Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitution: 1 X 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids. SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L-type amino acid transporter 3; Large neutral amino acids transporter small subunit 3; prostate cancer overexpressed gene 1; Prostate cancer overexpressed gene 1 protein; SLC43A1; solute carrier family 43 (amino acid system L transporter), member 1; Solute carrier family 43 member 1; solute carrier family 43, member 1
Gene Aliases: 2610016F07Rik; AA986141; LAT3; PB39; POV1; R00504; SLC43A1
UniProt ID: (Human) O75387, (Mouse) Q8BSM7
Entrez Gene ID: (Human) 8501, (Mouse) 72401, (Rat) 311168
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support