Product References
Denosumab Can Prevent Collapse in Patients with Early-Stage Steroid-Induced Osteonecrosis of the Femoral Head by Inhibiting Osteoclasts and Autophagy.
Orthopaedic surgery
Liu B,Gao F,Xiu X,Wu T,Liu Z,Zhang B,Liu S,Han Y
Sun Jan 01 00:00:00 UTC 2023
PSMD2 contributes to the progression of esophageal squamous cell carcinoma by repressing autophagy.
Cell & bioscience
Liu Y,Wu M,Xu S,Niu X,Liu W,Miao C,Lin A,Xu Y,Yu L
Thu Mar 30 00:00:00 UTC 2023
PSMD2 contributes to the progression of esophageal squamous cell carcinoma by repressing autophagy.
Cell & bioscience
Liu Y,Wu M,Xu S,Niu X,Liu W,Miao C,Lin A,Xu Y,Yu L
Thu Mar 30 00:00:00 UTC 2023
Buyang Huanwu Decoction alleviates cerebral ischemic injury through modulating caveolin-1-mediated mitochondrial quality control.
Frontiers in pharmacology
Xu Y,Chen B,Yi J,Tian F,Liu Y,Ouyang Y,Yuan C,Liu B
Sun May 28 00:00:00 UTC 2023
Rapamycin Inhibits Osteoclastogenesis and Prevents LPS-Induced Alveolar Bone Loss by Oxidative Stress Suppression.
ACS omega
Feng C,Liu Y,Zhang BY,Zhang H,Shan FY,Li TQ,Zhao ZN,Wang XX,Zhang XY
Tue Jun 13 00:00:00 UTC 2023
Rosuvastatin promotes survival of random skin flaps through AMPK-mTOR pathway-induced autophagy.
International immunopharmacology
Ye H,Li F,Shen Y,Wu X,Zhao L,Zhang H,Yang J,Shui X
Mon May 01 00:00:00 UTC 2023
Rosuvastatin promotes survival of random skin flaps through AMPK-mTOR pathway-induced autophagy.
International immunopharmacology
Ye H,Li F,Shen Y,Wu X,Zhao L,Zhang H,Yang J,Shui X
Mon May 01 00:00:00 UTC 2023
Resveratrol induces proliferation and differentiation of mouse pre-osteoblast MC3T3-E1 by promoting autophagy.
BMC complementary medicine and therapies
Cai W,Sun B,Song C,Liu F,Wu Z,Liu Z
Fri Apr 14 00:00:00 UTC 2023
Autophagy induced by Rab1a-ULK1 interaction promotes porcine reproductive and respiratory syndrome virus replication.
Virus research
Jiang C,Diao F,Ma Z,Zhang J,Bai J,Nauwynck H,Jiang P,Liu X
Mon Jan 02 00:00:00 UTC 2023
Physiological ischemic training improves cardiac function through the attenuation of cardiomyocyte apoptosis and the activation of the vagus nerve in chronic heart failure.
Frontiers in neuroscience
Zhu X,Wang S,Cheng Y,Gu H,Zhang X,Teng M,Zhang Y,Wang J,Hua W,Lu X
Tue May 09 00:00:00 UTC 2023
LncRNA LZTS1-AS1 induces proliferation, metastasis and inhibits autophagy of pancreatic cancer cells through the miR-532 /TWIST1 signaling pathway.
Cancer cell international
Wu H,Li A,Zheng Q,Gu J,Zhou W
Tue Jul 04 00:00:00 UTC 2023
ATB0,+-targeted nanoparticles initiate autophagy suppression to overcome chemoresistance for enhanced colorectal cancer therapy.
International journal of pharmaceutics
Chen Z,Chen H,Huang L,Duan B,Dai S,Cai W,Sun M,Jiang Z,Lu R,Jiang Y,Jiang X,Zheng H,Yao Q,Kim K,Lin G,Xie C,Chu M,Chen R,Kou L
Sun Jun 25 00:00:00 UTC 2023
Dioscin ameliorates silica-aggravated systemic lupus erythematosus via suppressing apoptosis and improving LC3-associated phagocytosis in MRL/lpr mice.
International immunopharmacology
Zhang P,Lei X,Ou L,Cheng Y,Miao Q,Li C,Chen Y
Wed Mar 01 00:00:00 UTC 2023
Autophagy-Related Gene WD Repeat Domain 45B Promotes Tumor Proliferation and Migration of Hepatocellular Carcinoma through the Akt/mTOR Signaling Pathway.
Diagnostics (Basel, Switzerland)
Li J,Chen L,Pang J,Yang C,Xie W,Shen G,Chen H,Li X,Xiao SY,Li Y
Mon Feb 27 00:00:00 UTC 2023
Osteogenic effects of rapamycin on bone marrow mesenchymal stem cells via inducing autophagy.
Journal of orthopaedic surgery and research
Xing Y,Liu C,Zhou L,Li Y,Wu D
Wed Feb 22 00:00:00 UTC 2023
Osteogenic effects of rapamycin on bone marrow mesenchymal stem cells via inducing autophagy.
Journal of orthopaedic surgery and research
Xing Y,Liu C,Zhou L,Li Y,Wu D
Wed Feb 22 00:00:00 UTC 2023
Purvalanol A induces apoptosis and reverses cisplatin resistance in ovarian cancer.
Anti-cancer drugs
Zhang X,Hong S,Yang J,Liu J,Wang Y,Peng J,Wang H,Hong L
Sun Jan 01 00:00:00 UTC 2023
Purvalanol A induces apoptosis and reverses cisplatin resistance in ovarian cancer.
Anti-cancer drugs
Zhang X,Hong S,Yang J,Liu J,Wang Y,Peng J,Wang H,Hong L
Sun Jan 01 00:00:00 UTC 2023
CircHIPK3 negatively regulates autophagy by blocking VCP binding to the Beclin 1 complex in bladder cancer.
Discover. Oncology
Wang C,Liu T,Wang J,Cheng C,Zhang Z,Zhang J,Huang H,Li Y
Sat Jun 03 00:00:00 UTC 2023
CXCR3 Inhibition Blocks the NF-κB Signaling Pathway by Elevating Autophagy to Ameliorate Lipopolysaccharide-Induced Intestinal Dysfunction in Mice.
Cells
Zhang C,Deng Y,Zhang Y,Ba T,Niu S,Chen Y,Gao Y,Dai H
Sun Jan 01 00:00:00 UTC 2023
CXCR3 Inhibition Blocks the NF-κB Signaling Pathway by Elevating Autophagy to Ameliorate Lipopolysaccharide-Induced Intestinal Dysfunction in Mice.
Cells
Zhang C,Deng Y,Zhang Y,Ba T,Niu S,Chen Y,Gao Y,Dai H
Sun Jan 01 00:00:00 UTC 2023
CXCR3 Inhibition Blocks the NF-κB Signaling Pathway by Elevating Autophagy to Ameliorate Lipopolysaccharide-Induced Intestinal Dysfunction in Mice.
Cells
Zhang C,Deng Y,Zhang Y,Ba T,Niu S,Chen Y,Gao Y,Dai H
Sun Jan 01 00:00:00 UTC 2023
CXCR3 Inhibition Blocks the NF-κB Signaling Pathway by Elevating Autophagy to Ameliorate Lipopolysaccharide-Induced Intestinal Dysfunction in Mice.
Cells
Zhang C,Deng Y,Zhang Y,Ba T,Niu S,Chen Y,Gao Y,Dai H
Sun Jan 01 00:00:00 UTC 2023
S-Doped carbonized polymer dots inhibit early myocardial fibrosis by regulating mitochondrial function.
Biomaterials science
Wang Y,Yang M,Zhang J,Ren J,Liu N,Liu B,Lu L,Yang B
Tue Jan 31 00:00:00 UTC 2023
Blue LED light induces cytotoxicity via ROS production and mitochondrial damage in bovine subcutaneous preadipocytes.
Environmental pollution (Barking, Essex : 1987)
Hao W,Zhao C,Li G,Wang H,Li T,Yan P,Wei S
Sat Apr 01 00:00:00 UTC 2023
PPARγ inhibited tumor immune escape by inducing PD-L1 autophagic degradation.
Cancer science
Gou Q,Che S,Chen M,Chen H,Shi J,Hou Y
Sat Jul 01 00:00:00 UTC 2023
Vitamin D Receptor Activation Targets ROS-Mediated Crosstalk Between Autophagy and Apoptosis in Hepatocytes in Cholestasic Mice.
Cellular and molecular gastroenterology and hepatology
Zheng Z,Xie J,Ma L,Hao Z,Zhang W,Li L
Tue Mar 21 00:00:00 UTC 2023
Sulforaphane prevents LPS-induced inflammation by regulating the Nrf2-mediated autophagy pathway in goat mammary epithelial cells and a mouse model of mastitis.
Journal of animal science and biotechnology
Shao D,Shen W,Miao Y,Gao Z,Pan M,Wei Q,Yan Z,Zhao X,Ma B
Wed May 03 00:00:00 UTC 2023
Sulforaphane prevents LPS-induced inflammation by regulating the Nrf2-mediated autophagy pathway in goat mammary epithelial cells and a mouse model of mastitis.
Journal of animal science and biotechnology
Shao D,Shen W,Miao Y,Gao Z,Pan M,Wei Q,Yan Z,Zhao X,Ma B
Wed May 03 00:00:00 UTC 2023
Sulforaphane prevents LPS-induced inflammation by regulating the Nrf2-mediated autophagy pathway in goat mammary epithelial cells and a mouse model of mastitis.
Journal of animal science and biotechnology
Shao D,Shen W,Miao Y,Gao Z,Pan M,Wei Q,Yan Z,Zhao X,Ma B
Wed May 03 00:00:00 UTC 2023
Sulforaphane prevents LPS-induced inflammation by regulating the Nrf2-mediated autophagy pathway in goat mammary epithelial cells and a mouse model of mastitis.
Journal of animal science and biotechnology
Shao D,Shen W,Miao Y,Gao Z,Pan M,Wei Q,Yan Z,Zhao X,Ma B
Wed May 03 00:00:00 UTC 2023
The adhesion G protein-coupled receptor VLGR1/ADGRV1 controls autophagy.
Basic & clinical pharmacology & toxicology
Linnert J,Güler BE,Krzysko J,Wolfrum U
Sun Oct 01 00:00:00 UTC 2023
Xiaojianzhong decoction prevents gastric precancerous lesions in rats by inhibiting autophagy and glycolysis in gastric mucosal cells.
World journal of gastrointestinal oncology
Zhang JX,Bao SC,Chen J,Chen T,Wei HL,Zhou XY,Li JT,Yan SG
Wed Mar 15 00:00:00 UTC 2023
The protective effect of inhibiting mitochondrial fission on the juvenile rat brain following PTZ kindling through inhibiting the BCL2L13/LC3 mitophagy pathway.
Metabolic brain disease
Fang Q,Zheng S,Chen Q,Chen L,Yang Y,Wang Y,Zhang H,Chen J
Wed Feb 01 00:00:00 UTC 2023
A Novel Phenazine Analog, CPUL1, Suppresses Autophagic Flux and Proliferation in Hepatocellular Carcinoma: Insight from Integrated Transcriptomic and Metabolomic Analysis.
Cancers
Chen J,Feng D,Lu Y,Zhang Y,Jiang H,Yuan M,Xu Y,Zou J,Zhu Y,Zhang J,Ge C,Wang Y
Sun Mar 05 00:00:00 UTC 2023
A Novel Phenazine Analog, CPUL1, Suppresses Autophagic Flux and Proliferation in Hepatocellular Carcinoma: Insight from Integrated Transcriptomic and Metabolomic Analysis.
Cancers
Chen J,Feng D,Lu Y,Zhang Y,Jiang H,Yuan M,Xu Y,Zou J,Zhu Y,Zhang J,Ge C,Wang Y
Sun Mar 05 00:00:00 UTC 2023
Tailor-Made Autophagy Cascade Amplification Polymeric Nanoparticles for Enhanced Tumor Immunotherapy.
Small (Weinheim an der Bergstrasse, Germany)
Long X,Wang H,Yan J,Li Y,Dong X,Tian S,Sun Y,Luo K,He B,Liang Y
Thu Jun 01 00:00:00 UTC 2023
BMAL1/p53 mediating bronchial epithelial cell autophagy contributes to PM2.5-aggravated asthma.
Cell communication and signaling : CCS
Chen SJ,Huang Y,Yu F,Feng X,Zheng YY,Li Q,Niu Q,Jiang YH,Zhao LQ,Wang M,Cheng PP,Song LJ,Liang LM,He XL,Xiong L,Xiang F,Wang X,Ma WL,Ye H
Mon Feb 20 00:00:00 UTC 2023
BMAL1/p53 mediating bronchial epithelial cell autophagy contributes to PM2.5-aggravated asthma.
Cell communication and signaling : CCS
Chen SJ,Huang Y,Yu F,Feng X,Zheng YY,Li Q,Niu Q,Jiang YH,Zhao LQ,Wang M,Cheng PP,Song LJ,Liang LM,He XL,Xiong L,Xiang F,Wang X,Ma WL,Ye H
Mon Feb 20 00:00:00 UTC 2023
BMAL1/p53 mediating bronchial epithelial cell autophagy contributes to PM2.5-aggravated asthma.
Cell communication and signaling : CCS
Chen SJ,Huang Y,Yu F,Feng X,Zheng YY,Li Q,Niu Q,Jiang YH,Zhao LQ,Wang M,Cheng PP,Song LJ,Liang LM,He XL,Xiong L,Xiang F,Wang X,Ma WL,Ye H
Mon Feb 20 00:00:00 UTC 2023
BMAL1/p53 mediating bronchial epithelial cell autophagy contributes to PM2.5-aggravated asthma.
Cell communication and signaling : CCS
Chen SJ,Huang Y,Yu F,Feng X,Zheng YY,Li Q,Niu Q,Jiang YH,Zhao LQ,Wang M,Cheng PP,Song LJ,Liang LM,He XL,Xiong L,Xiang F,Wang X,Ma WL,Ye H
Mon Feb 20 00:00:00 UTC 2023
Aa-Z2 triggers ROS-induced apoptosis of osteosarcoma by targeting PDK-1.
Journal of translational medicine
Liu Y,She W,Li Y,Wang M,Liu Y,Ning B,Xu T,Huang T,Wei Y
Sat Jan 07 00:00:00 UTC 2023
Stromal cell-derived small extracellular vesicles enhance radioresistance of prostate cancer cells via interleukin-8-induced autophagy.
Journal of extracellular vesicles
Wang X,Xu F,Kou H,Zheng Y,Yang J,Xu Z,Fang Y,Sun W,Zhu S,Jiang Q,Wei X,Xu Y
Sat Jul 01 00:00:00 UTC 2023
Stromal cell-derived small extracellular vesicles enhance radioresistance of prostate cancer cells via interleukin-8-induced autophagy.
Journal of extracellular vesicles
Wang X,Xu F,Kou H,Zheng Y,Yang J,Xu Z,Fang Y,Sun W,Zhu S,Jiang Q,Wei X,Xu Y
Sat Jul 01 00:00:00 UTC 2023
Sirt1 Overexpression Inhibits Fibrous Scar Formation and Improves Functional Recovery After Cerebral Ischemic Injury Through the Deacetylation of 14-3-3ζ.
Molecular neurobiology
Chen Y,Huang J,Liu J,Zhu H,Li X,Wen J,Tian M,Ren J,Zhou L,Yang Q
Tue Aug 01 00:00:00 UTC 2023
The role of Sirt3-induced autophagy in renal structural damage caused by periodontitis in rats.
Journal of periodontal research
Ding X,Hou Y,Liu X,Li X,Liu X,Deng Y,Cao N,Yu W
Wed Feb 01 00:00:00 UTC 2023
Combined effects of EMP and RF field on emotional behavior in mice.
Frontiers in public health
Qin T,Liu L,Wang X,Guo L,Lin J,Du J,Xue Y,Lai P,Jing Y,Ding G
Tue Apr 04 00:00:00 UTC 2023
Combined effects of EMP and RF field on emotional behavior in mice.
Frontiers in public health
Qin T,Liu L,Wang X,Guo L,Lin J,Du J,Xue Y,Lai P,Jing Y,Ding G
Tue Apr 04 00:00:00 UTC 2023
Deubiquitylase PSMD14 inhibits autophagy to promote ovarian cancer progression via stabilization of LRPPRC.
Biochimica et biophysica acta. Molecular basis of disease
Zhao Z,Xu H,Wei Y,Sun L,Song Y
Wed Feb 01 00:00:00 UTC 2023
Deubiquitylase PSMD14 inhibits autophagy to promote ovarian cancer progression via stabilization of LRPPRC.
Biochimica et biophysica acta. Molecular basis of disease
Zhao Z,Xu H,Wei Y,Sun L,Song Y
Wed Feb 01 00:00:00 UTC 2023
Naringin regulates endoplasmic reticulum stress and mitophagy through the ATF3/PINK1 signaling axis to alleviate pulmonary fibrosis.
Naunyn-Schmiedeberg's archives of pharmacology
Wei Y,Sun L,Liu C,Li L
Thu Jun 01 00:00:00 UTC 2023
PPARγ agonist pioglitazone enhances colorectal cancer immunotherapy by inducing PD-L1 autophagic degradation.
European journal of pharmacology
Jia X,Qian J,Chen H,Liu Q,Hussain S,Jin J,Shi J,Hou Y
Wed Jul 05 00:00:00 UTC 2023
Fluoride-Induced Cortical Toxicity in Rats: the Role of Excessive Endoplasmic Reticulum Stress and Its Mediated Defective Autophagy.
Biological trace element research
Zhang J,Tang Y,Xu W,Hu Z,Xu S,Niu Q
Tue Aug 01 00:00:00 UTC 2023
Fluoride-Induced Cortical Toxicity in Rats: the Role of Excessive Endoplasmic Reticulum Stress and Its Mediated Defective Autophagy.
Biological trace element research
Zhang J,Tang Y,Xu W,Hu Z,Xu S,Niu Q
Tue Aug 01 00:00:00 UTC 2023
Albumosomes formed by cytoplasmic pre-folding albumin maintain mitochondrial homeostasis and inhibit nonalcoholic fatty liver disease.
Signal transduction and targeted therapy
Ma B,Ju A,Zhang S,An Q,Xu S,Liu J,Yu L,Fu Y,Luo Y
Fri Jun 16 00:00:00 UTC 2023
A copper complex that combats triple negative breast cancer by restraining angiogenesis.
Dalton transactions (Cambridge, England : 2003)
Zheng H,Hu C,Quan Y,Ye X,Shi X,Guo Z,Wang X
Tue Jun 06 00:00:00 UTC 2023
A copper complex that combats triple negative breast cancer by restraining angiogenesis.
Dalton transactions (Cambridge, England : 2003)
Zheng H,Hu C,Quan Y,Ye X,Shi X,Guo Z,Wang X
Tue Jun 06 00:00:00 UTC 2023
ETNPPL impairs autophagy through regulation of the ARG2-ROS signaling axis, contributing to palmitic acid-induced hepatic insulin resistance.
Free radical biology & medicine
Wang C,Li X,Zhang W,Liu W,Lv Z,Gui R,Li M,Li Y,Sun X,Liu P,Fan X,Yang S,Xiong Y,Qian L
Sat Apr 01 00:00:00 UTC 2023
Nesfatin-1 suppresses autophagy of chondrocytes in osteoarthritis via remodeling of cytoskeleton and inhibiting RhoA/ROCK signal pathway.
Journal of orthopaedic surgery and research
Jiang L,Moqbel SAA,Zhu J,Fu Q,Lai J,Lin C,Wu L
Thu Mar 02 00:00:00 UTC 2023
Nesfatin-1 suppresses autophagy of chondrocytes in osteoarthritis via remodeling of cytoskeleton and inhibiting RhoA/ROCK signal pathway.
Journal of orthopaedic surgery and research
Jiang L,Moqbel SAA,Zhu J,Fu Q,Lai J,Lin C,Wu L
Thu Mar 02 00:00:00 UTC 2023
Discovery of rafoxanide as a novel agent for the treatment of non-small cell lung cancer.
Scientific reports
Hu A,Liu J,Wang Y,Zhang M,Guo Y,Qin Y,Liu T,Men Y,Chen Q,Liu T
Fri Jan 13 00:00:00 UTC 2023
Discovery of rafoxanide as a novel agent for the treatment of non-small cell lung cancer.
Scientific reports
Hu A,Liu J,Wang Y,Zhang M,Guo Y,Qin Y,Liu T,Men Y,Chen Q,Liu T
Fri Jan 13 00:00:00 UTC 2023
Exendin-4 alleviates myocardial ischemia reperfusion injury by enhancing autophagy through promoting nuclear translocation of TFEB.
Experimental cell research
He W,Tong G,Fan H,Zhen C,Zeng L,Xue L,Chen J,Sun Z,He P
Wed Feb 15 00:00:00 UTC 2023
DBP induced autophagy and necrotic apoptosis in HepG2 cells via the mitochondrial damage pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Liu H,Xu L,Zhou L,Han W,Li Z,Liu C
Thu Jun 01 00:00:00 UTC 2023
Autophagy inhibition and ferroptosis activation during atherosclerosis: Hypoxia-inducible factor 1α inhibitor PX-478 alleviates atherosclerosis by inducing autophagy and suppressing ferroptosis in macrophages.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Hu G,Yuan Z,Wang J
Mon May 01 00:00:00 UTC 2023
Autophagy inhibition and ferroptosis activation during atherosclerosis: Hypoxia-inducible factor 1α inhibitor PX-478 alleviates atherosclerosis by inducing autophagy and suppressing ferroptosis in macrophages.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Hu G,Yuan Z,Wang J
Mon May 01 00:00:00 UTC 2023
BNIP3-mediated autophagy via the mTOR/ULK1 pathway induces primordial follicle loss after ovarian tissue transplantation.
Journal of assisted reproduction and genetics
Liu F,Li M
Wed Mar 01 00:00:00 UTC 2023
BNIP3-mediated autophagy via the mTOR/ULK1 pathway induces primordial follicle loss after ovarian tissue transplantation.
Journal of assisted reproduction and genetics
Liu F,Li M
Wed Mar 01 00:00:00 UTC 2023
Dexmedetomidine alleviates cerebral ischemia-reperfusion injury via inhibiting autophagy through PI3K/Akt/mTOR pathway.
Journal of molecular histology
Li J,Wang K,Liu M,He J,Zhang H,Liu H
Thu Jun 01 00:00:00 UTC 2023
Autophagy-Targeted Calcium Phosphate Nanoparticles Enable Transarterial Chemoembolization for Enhanced Cancer Therapy.
ACS applied materials & interfaces
Yuan G,Xu Y,Bai X,Wang W,Wu X,Chen J,Li J,Jia X,Gu Z,Zhang X,Hu W,Wang J,Liu Y,Zhu XM
Wed Mar 08 00:00:00 UTC 2023
Subacute ruminal acidosis downregulates FOXA2, changes oxidative status, and induces autophagy in the livers of dairy cows fed a high-concentrate diet.
Journal of dairy science
Zhang H,Xue Y,Xie W,Wang Y,Ma N,Chang G,Shen X
Wed Mar 01 00:00:00 UTC 2023
P4HA2-mediated HIF-1α stabilization promotes erdafitinib-resistance in FGFR3-alteration bladder cancer.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li X,Li Y,Liu B,Chen L,Lyu F,Zhang P,He Q,Cheng L,Liu C,Song Y,Xing Y
Sat Apr 01 00:00:00 UTC 2023
Follicular fluid exosomes regulate OVGP1 secretion in yak oviduct epithelial cells via autophagy in vitro.
Journal of cellular physiology
Wang J,Pan Y,Wang M,Xu R,Han X,Ma R,Zhao L,Zhang T,Wang Y,Zhao T,Ding T,Baloch AR,Wang L,Cui Y,Yu S
Mon May 01 00:00:00 UTC 2023
Molecular mechanism by which the Notch signaling pathway regulates autophagy in a rat model of pulmonary fibrosis in pigeon breeder's lung.
Open medicine (Warsaw, Poland)
Li Y,Lian Z,Li Q,Ding W,Wang W,Zhang L,Muhataer X,Zhou Y,Yang X,Wu C
Wed Feb 15 00:00:00 UTC 2023
Micelle encapsulation zinc-doped copper oxide nanocomposites reverse Olaparib resistance in ovarian cancer by disrupting homologous recombination repair.
Bioengineering & translational medicine
Yi J,Luo X,Xing J,Gedanken A,Lin X,Zhang C,Qiao G
Mon May 01 00:00:00 UTC 2023
Mesenchymal stem cell-derived extracellular vesicles carrying miR-99b-3p restrain microglial activation and neuropathic pain by stimulating autophagy.
International immunopharmacology
Gao X,Gao LF,Kong XQ,Zhang YN,Jia S,Meng CY
Wed Feb 01 00:00:00 UTC 2023
Inhibition of PINK1-Mediated Mitophagy Contributes to Postoperative Cognitive Dysfunction through Activation of Caspase-3/GSDME-Dependent Pyroptosis.
ACS chemical neuroscience
Wang W,Zhao B,Gao W,Song W,Hou J,Zhang L,Xia Z
Wed Apr 05 00:00:00 UTC 2023
Mitochondrial pyruvate carrier 1 alleviates hypoxic-ischemic brain injury in rats.
Life sciences
Liu Y,Yuan Y,Yan Y,Wang R,Wang Z,Liu X,Zhang Y,Hua J,Wang Y,Zhao L
Sat Jul 15 00:00:00 UTC 2023
Electroacupuncture repairs intestinal barrier by upregulating CB1 through gut microbiota in DSS-induced acute colitis.
Chinese medicine
Yang J,Wang L,Mei M,Guo J,Yang X,Liu S
Fri Mar 10 00:00:00 UTC 2023
Protective effects of melatonin on deoxynivalenol-induced oxidative stress and autophagy in IPEC-J2 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Xu Y,Xie Y,Wu Z,Wang H,Chen Z,Wang J,Bao W
Sat Jul 01 00:00:00 UTC 2023
Exosomes derived from BMSCs ameliorate cyclophosphamide-induced testosterone deficiency by enhancing the autophagy of Leydig cells via the AMPK-mTOR signaling pathway.
Asian journal of andrology
Liang HY,Peng F,Pan MJ,Liao SL,Wei C,Wei GY,Xie X,Xue KY,Chen MK,Yang JK,Guo WB,Liu CD,Zhou QZ
Fri Jul 21 00:00:00 UTC 2023
Newly identified lncRNA-45 promotes breast cancer metastasis through activating the mTOR signaling pathway.
Biochemical and biophysical research communications
Qiu J,Guo Y,Wang S,Ren Q,Dong Z,Gao M,Ma J,Chen S,Liu S
Sun Jan 15 00:00:00 UTC 2023
Ganoapplins A and B with an unprecedented 6/6/6/5/6-fused pentacyclic skeleton from Ganoderma inhibit Tau pathology through activating autophagy.
Bioorganic chemistry
Peng X,Luo R,Ran X,Guo Y,Yao YG,Qiu M
Wed Mar 01 00:00:00 UTC 2023
Necroptosis signaling and mitochondrial dysfunction cross-talking facilitate cell death mediated by chelerythrine in glioma.
Free radical biology & medicine
Wang P,Zheng SY,Jiang RL,Wu HD,Li YA,Lu JL,Xiong Y,Han B,Lin L
Thu Jun 01 00:00:00 UTC 2023
EFFECT OF EXOSOMES DERIVED FROM BONE MARROW MESENCHYMAL STEM CELLS ON PROGRAMMED CELL DEATH IN BLAST-INDUCED LUNG INJURY IN RATS.
Shock (Augusta, Ga.)
Deng M,He J,Hao C,Guo Y,Si H,Deng X,Zhang C,Li S,Yao S,Ren W,Yao W
Thu Jun 01 00:00:00 UTC 2023
Autophagic response of intestinal epithelial cells exposed to polystyrene nanoplastics.
Environmental toxicology
Xu X,Feng Y,Han C,Yao Z,Liu Y,Luo C,Sheng J
Sun Jan 01 00:00:00 UTC 2023
Autophagic response of intestinal epithelial cells exposed to polystyrene nanoplastics.
Environmental toxicology
Xu X,Feng Y,Han C,Yao Z,Liu Y,Luo C,Sheng J
Sun Jan 01 00:00:00 UTC 2023
Autophagic response of intestinal epithelial cells exposed to polystyrene nanoplastics.
Environmental toxicology
Xu X,Feng Y,Han C,Yao Z,Liu Y,Luo C,Sheng J
Sun Jan 01 00:00:00 UTC 2023
Autophagic response of intestinal epithelial cells exposed to polystyrene nanoplastics.
Environmental toxicology
Xu X,Feng Y,Han C,Yao Z,Liu Y,Luo C,Sheng J
Sun Jan 01 00:00:00 UTC 2023
Cholesterol homeostasis regulated by ABCA1 is critical for retinal ganglion cell survival.
Science China. Life sciences
Yang J,Chen Y,Zou T,Xue B,Yang F,Wang X,Huo Y,Yan B,Xu Y,He S,Yin Y,Wang J,Zhu X,Zhang L,Zhou Y,Tai Z,Shuai P,Yu M,Luo Q,Cheng Y,Gong B,Zhu X,Zhang J,Sun X,Lin Y,Zhang H,Yang Z
Wed Feb 01 00:00:00 UTC 2023
Cholesterol homeostasis regulated by ABCA1 is critical for retinal ganglion cell survival.
Science China. Life sciences
Yang J,Chen Y,Zou T,Xue B,Yang F,Wang X,Huo Y,Yan B,Xu Y,He S,Yin Y,Wang J,Zhu X,Zhang L,Zhou Y,Tai Z,Shuai P,Yu M,Luo Q,Cheng Y,Gong B,Zhu X,Zhang J,Sun X,Lin Y,Zhang H,Yang Z
Wed Feb 01 00:00:00 UTC 2023
Promotion of colorectal cancer cell death by ezetimibe via mTOR signaling-dependent mitochondrial dysfunction.
Frontiers in pharmacology
Zheng Y,Yang W,Jia Y,Ji J,Wu L,Feng J,Li Y,Cheng Z,Zhang J,Li J,Dai W,Xu X,Wu J,Zhou Y,Guo C
Tue Feb 28 00:00:00 UTC 2023
mTOR/ULK1 signaling axis mediated ultrashort wave regulation of autophagy to alleviate diabetic kidney disease.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University
Yan Y,Yu S,Fan Y
Wed Nov 01 00:00:00 UTC 2023
Active post-transcriptional regulation and ACLY-mediated acetyl-CoA synthesis as a pivotal target of Shuang-Huang-Sheng-Bai formula for lung adenocarcinoma treatment.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu D,Dong C,Wang F,Liu W,Jin X,Qi SL,Liu L,Jin Q,Wang S,Wu J,Wang C,Yang J,Deng H,Cai Y,Yang L,Qin J,Zhang C,Yang X,Wang MS,Yu G,Xue YW,Wang Z,Ge GB,Xu Z,Chen WL
Mon May 01 00:00:00 UTC 2023
NKAP acts with HDAC3 to prevent R-loop associated genome instability.
Cell death and differentiation
Zhang X,Duan J,Li Y,Jin X,Wu C,Yang X,Lu W,Ge W
Sat Jul 01 00:00:00 UTC 2023
HDAC inhibitor chidamide overcomes drug resistance in chronic myeloid leukemia with the T315i mutation through the Akt-autophagy pathway.
Human cell
Yin L,Zhang Q,Xie S,Cheng Z,Li R,Zhu H,Yu Q,Yuan H,Wang C,Peng H,Zhang G
Sat Jul 01 00:00:00 UTC 2023
Golgi damage caused by dysfunction of PiT-2 in primary familial brain calcification.
Biochemical and biophysical research communications
Sun H,Wang Z,Zhang Q,Chen N,Tang M,Yang Z,Xu Y,Kang J,Wang Y
Sun Jan 29 00:00:00 UTC 2023
The inhibition of TRPML1/TFEB leads to lysosomal biogenesis disorder, contributes to developmental fluoride neurotoxicity.
Ecotoxicology and environmental safety
Zhang J,Tang Y,Hu Z,Xu W,Ma Y,Xu P,Xing H,Niu Q
Sun Jan 15 00:00:00 UTC 2023
Induction of Sestrin2 by pterostilbene suppresses ethanol-triggered hepatocyte senescence by degrading CCN1 via p62-dependent selective autophagy.
Cell biology and toxicology
Jiang Y,Zhou Y,Xu W,Wang X,Jin H,Bao X,Lu C
Thu Jun 01 00:00:00 UTC 2023
Xue-Jie-San prevents the early development of colitis-associated intestinal fibrosis by blocking Notch1 and FGL1 signaling pathways.
Journal of ethnopharmacology
Gao Y,Lu LJ,Zhang ZZ,Yang X,Du J,Wen K,Huang H,Wang XP,Sun XL
Sat Oct 28 00:00:00 UTC 2023
Xue-Jie-San prevents the early development of colitis-associated intestinal fibrosis by blocking Notch1 and FGL1 signaling pathways.
Journal of ethnopharmacology
Gao Y,Lu LJ,Zhang ZZ,Yang X,Du J,Wen K,Huang H,Wang XP,Sun XL
Sat Oct 28 00:00:00 UTC 2023
Analgesic Effect of Perineural Injection of BoNT/A on Neuropathic Pain Induced by Chronic Constriction Injury of Sciatic Nerve in Rats.
Neurochemical research
He JJ,Wei XM,Wu ML,Song ZB,Jiang L,Zhang WX
Sat Jul 01 00:00:00 UTC 2023
RBM3 interacts with Raptor to regulate autophagy and protect cardiomyocytes from ischemia-reperfusion-induced injury.
Journal of physiology and biochemistry
Wang N,Wang L,Li C,Rao P,Wang X,Xu Y,Yang L,Sun L,Lu D
Wed Feb 01 00:00:00 UTC 2023
RBM3 interacts with Raptor to regulate autophagy and protect cardiomyocytes from ischemia-reperfusion-induced injury.
Journal of physiology and biochemistry
Wang N,Wang L,Li C,Rao P,Wang X,Xu Y,Yang L,Sun L,Lu D
Wed Feb 01 00:00:00 UTC 2023
Combination of DNA Damage, Autophagy, and ERK Inhibition: Novel Evodiamine-Inspired Multi-Action Pt(IV) Prodrugs with High-Efficiency and Low-Toxicity Antitumor Activity.
Journal of medicinal chemistry
Liu XM,Li Z,Xie XR,Wang JQ,Qiao X,Qiao X,Xie CZ,Xu JY
Thu Feb 09 00:00:00 UTC 2023
Combination of DNA Damage, Autophagy, and ERK Inhibition: Novel Evodiamine-Inspired Multi-Action Pt(IV) Prodrugs with High-Efficiency and Low-Toxicity Antitumor Activity.
Journal of medicinal chemistry
Liu XM,Li Z,Xie XR,Wang JQ,Qiao X,Qiao X,Xie CZ,Xu JY
Thu Feb 09 00:00:00 UTC 2023
Novel 2-Amino-1,4-Naphthoquinone Derivatives Induce A549 Cell Death through Autophagy.
Molecules (Basel, Switzerland)
Tan HY,Liang FM,Zhang WJ,Zhang Y,Cui JH,Dai YY,Qiu XM,Wang WH,Zhou Y,Chen DP,Li CP
Fri Apr 07 00:00:00 UTC 2023
AmAtg2B-Mediated Lipophagy Regulates Lipolysis of Pupae in Apis mellifera.
International journal of molecular sciences
Chen WF,Wang HF,Wang Y,Liu ZG,Xu BH
Fri Jan 20 00:00:00 UTC 2023
AmAtg2B-Mediated Lipophagy Regulates Lipolysis of Pupae in Apis mellifera.
International journal of molecular sciences
Chen WF,Wang HF,Wang Y,Liu ZG,Xu BH
Fri Jan 20 00:00:00 UTC 2023
Euphejolkinolide A, a new ent-abietane lactone from Euphorbia peplus L. with promising biological activity in activating the autophagy-lysosomal pathway.
Heliyon
Ran X,Lu QY,Li YY,Pu XX,Guo Y,Yuan MR,Guan SP,Sun M,Jiao L,Yao YG,Di YT,Hao XJ,Luo R
Wed Feb 01 00:00:00 UTC 2023
Iso-seco-tanapartholide induces p62 covalent oligomerization to activate KEAP1-NRF2 redox pathway in rheumatoid arthritis.
International immunopharmacology
Zhu D,Kong M,Chen C,Luo J,Kong L
Wed Feb 01 00:00:00 UTC 2023
Robo2 promotes osteoblast differentiation and mineralization through autophagy and is activated by parathyroid hormone induction.
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft
Yuan N,Wang X,He M
Thu Jun 01 00:00:00 UTC 2023
TRIM7/RNF90 promotes autophagy via regulation of ATG7 ubiquitination during L. monocytogenes infection.
Autophagy
Wang J,Qin X,Huang Y,Zhang Q,Pei J,Wang Y,Goren I,Ma S,Song Z,Liu Y,Xing H,Wang H,Yang B
Thu Jun 01 00:00:00 UTC 2023
TRIM7/RNF90 promotes autophagy via regulation of ATG7 ubiquitination during L. monocytogenes infection.
Autophagy
Wang J,Qin X,Huang Y,Zhang Q,Pei J,Wang Y,Goren I,Ma S,Song Z,Liu Y,Xing H,Wang H,Yang B
Thu Jun 01 00:00:00 UTC 2023
TRIM7/RNF90 promotes autophagy via regulation of ATG7 ubiquitination during L. monocytogenes infection.
Autophagy
Wang J,Qin X,Huang Y,Zhang Q,Pei J,Wang Y,Goren I,Ma S,Song Z,Liu Y,Xing H,Wang H,Yang B
Thu Jun 01 00:00:00 UTC 2023
TRIM7/RNF90 promotes autophagy via regulation of ATG7 ubiquitination during L. monocytogenes infection.
Autophagy
Wang J,Qin X,Huang Y,Zhang Q,Pei J,Wang Y,Goren I,Ma S,Song Z,Liu Y,Xing H,Wang H,Yang B
Thu Jun 01 00:00:00 UTC 2023
Treponema pallidum membrane protein Tp47 promotes angiogenesis through ROS-induced autophagy.
Journal of the European Academy of Dermatology and Venereology : JEADV
Li W,Xie L,Li QL,Xu QY,Lin LR,Liu LL,Yang TC
Wed Mar 01 00:00:00 UTC 2023
Cross-talk between autophagy and ferroptosis contributes to the liver injury induced by fluoride via the mtROS-dependent pathway.
Ecotoxicology and environmental safety
Xu W,Hu Z,Zhang J,Tang Y,Xing H,Xu P,Ma Y,Niu Q
Sun Jan 15 00:00:00 UTC 2023
HCMV-encoded viral protein US12 promotes autophagy by inducing autophagy flux.
Biochemical and biophysical research communications
Kim HJ,Lee Y,Lee S,Park B
Sun Apr 30 00:00:00 UTC 2023
HCMV-encoded viral protein US12 promotes autophagy by inducing autophagy flux.
Biochemical and biophysical research communications
Kim HJ,Lee Y,Lee S,Park B
Sun Apr 30 00:00:00 UTC 2023
Inhibition of TRF2 Leads to Ferroptosis, Autophagic Death, and Apoptosis by Causing Telomere Dysfunction.
Oxidative medicine and cellular longevity
Yang Q,Nie Z,Zhu Y,Hao M,Liu S,Ding X,Wang F,Wang F,Geng X
Wed May 03 00:00:00 UTC 2023
Role of omentin-1 in susceptibility to anxiety and depression like behaviors.
Molecular and cellular endocrinology
Ji L,Zhang L,Liang Z,Zhong S,Liu X,Liu Z,Poon WS,Song Y,Chen B,Wang R
Sun Aug 20 00:00:00 UTC 2023
Role of omentin-1 in susceptibility to anxiety and depression like behaviors.
Molecular and cellular endocrinology
Ji L,Zhang L,Liang Z,Zhong S,Liu X,Liu Z,Poon WS,Song Y,Chen B,Wang R
Sun Aug 20 00:00:00 UTC 2023
Enhanced autophagy reversed aflatoxin B1-induced decrease in lactate secretion of dairy goat Sertoli cells.
Ecotoxicology and environmental safety
Han S,Zhang H,Liu X,Wen F,Li B,Bie Z,Qiu H,Hu J
Sat Jul 01 00:00:00 UTC 2023
CircEPS15, as a sponge of MIR24-3p ameliorates neuronal damage in Parkinson disease through boosting PINK1-PRKN-mediated mitophagy.
Autophagy
Zhou Y,Liu Y,Kang Z,Yao H,Song N,Wang M,Song C,Zhang K,Ding J,Tang J,Hu G,Lu M
Fri Sep 01 00:00:00 UTC 2023
Ferredoxin 1 regulates granulosa cell apoptosis and autophagy in polycystic ovary syndrome.
Clinical science (London, England : 1979)
Xing J,Qiao G,Luo X,Liu S,Chen S,Ye G,Zhang C,Yi J
Fri Mar 31 00:00:00 UTC 2023
Tegument protein UL21 of alpha-herpesvirus inhibits the innate immunity by triggering CGAS degradation through TOLLIP-mediated selective autophagy.
Autophagy
Ma Z,Bai J,Jiang C,Zhu H,Liu D,Pan M,Wang X,Pi J,Jiang P,Liu X
Mon May 01 00:00:00 UTC 2023
Non-Lethal Concentrations of CdCl2 Cause Marked Alternations in Cellular Stress Responses within Exposed Sertoli Cell Line.
Toxics
Man Y,Liu Y,Xiong C,Zhang Y,Zhang L
Thu Feb 09 00:00:00 UTC 2023
Melatonin protects against developmental PBDE-47 neurotoxicity by targeting the AMPK/mitophagy axis.
Journal of pineal research
Dong L,Sun Q,Qiu H,Yang K,Xiao B,Xia T,Wang A,Gao H,Zhang S
Tue Aug 01 00:00:00 UTC 2023
Bone marrow-derived mesenchymal stem cell-conditioned medium ameliorates diabetic foot ulcers in rats.
Clinics (Sao Paulo, Brazil)
Xu YF,Wu YX,Wang HM,Gao CH,Xu YY,Yan Y
Thu Mar 30 00:00:00 UTC 2023
Apigenin Alleviated High-Fat-Diet-Induced Hepatic Pyroptosis by Mitophagy-ROS-CTSB-NLRP3 Pathway in Mice and AML12 Cells.
Journal of agricultural and food chemistry
Meng Z,Gao M,Wang C,Guan S,Zhang D,Lu J
Wed May 10 00:00:00 UTC 2023
The outer mitochondrial membrane protein TMEM11 demarcates spatially restricted BNIP3/BNIP3L-mediated mitophagy.
The Journal of cell biology
Gok MO,Connor OM,Wang X,Menezes CJ,Llamas CB,Mishra P,Friedman JR
Mon Apr 03 00:00:00 UTC 2023
Autophagy-mediated circHIPK2 promotes lipopolysaccharide-induced astrocytic inflammation via SIGMAR1.
International immunopharmacology
Huang R,Cai L,Ma X,Shen K
Sat Apr 01 00:00:00 UTC 2023
Porcine reproductive and respiratory syndrome virus infection triggers autophagy via ER stress-induced calcium signaling to facilitate virus replication.
PLoS pathogens
Diao F,Jiang C,Sun Y,Gao Y,Bai J,Nauwynck H,Wang X,Yang Y,Jiang P,Liu X
Wed Mar 01 00:00:00 UTC 2023
Porcine reproductive and respiratory syndrome virus infection triggers autophagy via ER stress-induced calcium signaling to facilitate virus replication.
PLoS pathogens
Diao F,Jiang C,Sun Y,Gao Y,Bai J,Nauwynck H,Wang X,Yang Y,Jiang P,Liu X
Wed Mar 01 00:00:00 UTC 2023
Hernandezine induces autophagic cell death in human pancreatic cancer cells via activation of the ROS/AMPK signaling pathway.
Acta pharmacologica Sinica
Song CF,Hu YH,Mang ZG,Ye Z,Chen HD,Jing DS,Fan GX,Ji SR,Yu XJ,Xu XW,Qin Y
Sat Apr 01 00:00:00 UTC 2023
Ursolic acid nanoparticles for glioblastoma therapy.
Nanomedicine : nanotechnology, biology, and medicine
Li Y,Zhao L,Zhao Q,Zhou Y,Zhou L,Song P,Liu B,Chen Q,Deng G
Thu Jun 01 00:00:00 UTC 2023
MLKL deficiency attenuated hepatocyte oxidative DNA damage by activating mitophagy to suppress macrophage cGAS-STING signaling during liver ischemia and reperfusion injury.
Cell death discovery
Xu J,Wu D,Zhou S,Hu H,Li F,Guan Z,Zhan X,Gao Y,Wang P,Rao Z
Fri Feb 10 00:00:00 UTC 2023
The organic arsenical-derived thioredoxin and glutathione system inhibitor ACZ2 induces apoptosis and autophagy in gastric cancer via ROS-dependent ER stress.
Biochemical pharmacology
Li Y,She W,Guo T,Huang T,Liu Y,Liu P,Xu X,Wang X,Wang M,Yu C,Liu Y,Wei Y
Wed Feb 01 00:00:00 UTC 2023
GSTP1-mediated S-glutathionylation of Pik3r1 is a redox hub that inhibits osteoclastogenesis through regulating autophagic flux.
Redox biology
Ji X,Hong J,Yang W,Yao M,Wang J,Jiang G,Wang Y,Li C,Lin J,Mou H,Li C,Li S,Chen Y,Shi M,Wang W,Lu F,Wu H,Zhao X,Qi Y,Yan S
Mon May 01 00:00:00 UTC 2023
Placental Mesenchymal Stem Cells Alleviate Podocyte Injury in Diabetic Kidney Disease by Modulating Mitophagy via the SIRT1-PGC-1alpha-TFAM Pathway.
International journal of molecular sciences
Han X,Wang J,Li R,Huang M,Yue G,Guan L,Deng Y,Cai W,Xu J
Tue Feb 28 00:00:00 UTC 2023
Placental Mesenchymal Stem Cells Alleviate Podocyte Injury in Diabetic Kidney Disease by Modulating Mitophagy via the SIRT1-PGC-1alpha-TFAM Pathway.
International journal of molecular sciences
Han X,Wang J,Li R,Huang M,Yue G,Guan L,Deng Y,Cai W,Xu J
Tue Feb 28 00:00:00 UTC 2023
Placental Mesenchymal Stem Cells Alleviate Podocyte Injury in Diabetic Kidney Disease by Modulating Mitophagy via the SIRT1-PGC-1alpha-TFAM Pathway.
International journal of molecular sciences
Han X,Wang J,Li R,Huang M,Yue G,Guan L,Deng Y,Cai W,Xu J
Tue Feb 28 00:00:00 UTC 2023
Placental Mesenchymal Stem Cells Alleviate Podocyte Injury in Diabetic Kidney Disease by Modulating Mitophagy via the SIRT1-PGC-1alpha-TFAM Pathway.
International journal of molecular sciences
Han X,Wang J,Li R,Huang M,Yue G,Guan L,Deng Y,Cai W,Xu J
Tue Feb 28 00:00:00 UTC 2023
Endoplasmic reticulum stress promotes sepsis-induced muscle atrophy via activation of STAT3 and Smad3.
Journal of cellular physiology
Zheng Y,Dai H,Chen R,Zhong Y,Zhou C,Wang Y,Zhan C,Luo J
Wed Mar 01 00:00:00 UTC 2023
Inhibitory Effects of Esculetin on Liver Cancer Through Triggering NCOA4 Pathway-Mediation Ferritinophagy in vivo and in vitro.
Journal of hepatocellular carcinoma
Xiu Z,Li Y,Fang J,Han J,Li S,Li Y,Yang X,Song G,Li Y,Jin N,Zhu Y,Zhu G,Sun L,Li X
Wed Apr 19 00:00:00 UTC 2023
Inhibitory Effects of Esculetin on Liver Cancer Through Triggering NCOA4 Pathway-Mediation Ferritinophagy in vivo and in vitro.
Journal of hepatocellular carcinoma
Xiu Z,Li Y,Fang J,Han J,Li S,Li Y,Yang X,Song G,Li Y,Jin N,Zhu Y,Zhu G,Sun L,Li X
Wed Apr 19 00:00:00 UTC 2023
Inhibitory Effects of Esculetin on Liver Cancer Through Triggering NCOA4 Pathway-Mediation Ferritinophagy in vivo and in vitro.
Journal of hepatocellular carcinoma
Xiu Z,Li Y,Fang J,Han J,Li S,Li Y,Yang X,Song G,Li Y,Jin N,Zhu Y,Zhu G,Sun L,Li X
Wed Apr 19 00:00:00 UTC 2023
Inhibitory Effects of Esculetin on Liver Cancer Through Triggering NCOA4 Pathway-Mediation Ferritinophagy in vivo and in vitro.
Journal of hepatocellular carcinoma
Xiu Z,Li Y,Fang J,Han J,Li S,Li Y,Yang X,Song G,Li Y,Jin N,Zhu Y,Zhu G,Sun L,Li X
Wed Apr 19 00:00:00 UTC 2023
Autophagy attenuates osteoarthritis in mice by inhibiting chondrocyte pyroptosis and improving subchondral bone remodeling.
Biomolecules & biomedicine
Yan J,Feng G,Yang Y,Ding D,Ma L,Zhao X,Chen X,Wang H,Chen Z,Jin Q
Wed Feb 01 00:00:00 UTC 2023
Diet-induced obesity augments ischemic myopathy and functional decline in a murine model of peripheral artery disease.
Translational research : the journal of laboratory and clinical medicine
Fletcher E,Miserlis D,Sorokolet K,Wilburn D,Bradley C,Papoutsi E,Wilkinson T,Ring A,Ferrer L,Haynatzki G,Smith RS,Bohannon WT,Koutakis P
Sun Oct 01 00:00:00 UTC 2023
Collateral activity of the CRISPR/RfxCas13d system in human cells.
Communications biology
Shi P,Murphy MR,Aparicio AO,Kesner JS,Fang Z,Chen Z,Trehan A,Guo Y,Wu X
Tue Mar 28 00:00:00 UTC 2023
DANCR promotes glioma cell autophagy and proliferation via the miR‑33b/DLX6/ATG7 axis.
Oncology reports
Yu W,Ma L,Li X
Wed Feb 01 00:00:00 UTC 2023
Zinc-Deficient Diet Causes Imbalance in Zinc Homeostasis and Impaired Autophagy and Impairs Semen Quality in Mice.
Biological trace element research
Sun B,Ma J,Te L,Zuo X,Liu J,Li Y,Bi J,Wang S
Mon May 01 00:00:00 UTC 2023
Gomphrena globosa L. extract alleviates carbon tetrachoride-induced liver injury in mice by activating antioxidant signaling pathways and promoting autophagy.
Molecular biology reports
Wei Y,Tan W,Qin H,Jin M,Yu H,Cheng J,Feng H
Sun Jan 01 00:00:00 UTC 2023
Hypoxia-induced GPCPD1 depalmitoylation triggers mitophagy via regulating PRKN-mediated ubiquitination of VDAC1.
Autophagy
Liu Y,Zhang H,Liu Y,Zhang S,Su P,Wang L,Li Y,Liang Y,Wang X,Zhao W,Chen B,Luo D,Zhang N,Yang Q
Fri Sep 01 00:00:00 UTC 2023
Chloroquine is a safe autophagy inhibitor for sustaining the expression of antioxidant enzymes in trophoblasts.
Journal of reproductive immunology
Furuta A,Shima T,Yoshida-Kawaguchi M,Yamada K,Yasuda I,Tsuda S,Yamaki-Ushijima A,Yoneda S,Higashisaka K,Cheng SB,Matsumoto K,Tsutsumi Y,Sharma S,Saito S,Nakashima A
Wed Feb 01 00:00:00 UTC 2023
Mesenchymal stromal cells ameliorate diabetes-induced muscle atrophy through exosomes by enhancing AMPK/ULK1-mediated autophagy.
Journal of cachexia, sarcopenia and muscle
Song J,Liu J,Cui C,Hu H,Zang N,Yang M,Yang J,Zou Y,Li J,Wang L,He Q,Guo X,Zhao R,Yan F,Liu F,Hou X,Sun Z,Chen L
Sat Apr 01 00:00:00 UTC 2023
Bone microenvironment regulative hydrogels with ROS scavenging and prolonged oxygen-generating for enhancing bone repair.
Bioactive materials
Sun H,Xu J,Wang Y,Shen S,Xu X,Zhang L,Jiang Q
Thu Jun 01 00:00:00 UTC 2023
Bone microenvironment regulative hydrogels with ROS scavenging and prolonged oxygen-generating for enhancing bone repair.
Bioactive materials
Sun H,Xu J,Wang Y,Shen S,Xu X,Zhang L,Jiang Q
Thu Jun 01 00:00:00 UTC 2023
New fungal protein from Pleurotus ferulae lanzi induces AMPK-mediated autophagy and G1-phase cell cycle arrest in A549 lung cancer cells.
International journal of biological macromolecules
Liu MH,Liu F,Ng TB,Liu ZK
Mon Jul 31 00:00:00 UTC 2023
New fungal protein from Pleurotus ferulae lanzi induces AMPK-mediated autophagy and G1-phase cell cycle arrest in A549 lung cancer cells.
International journal of biological macromolecules
Liu MH,Liu F,Ng TB,Liu ZK
Mon Jul 31 00:00:00 UTC 2023
New fungal protein from Pleurotus ferulae lanzi induces AMPK-mediated autophagy and G1-phase cell cycle arrest in A549 lung cancer cells.
International journal of biological macromolecules
Liu MH,Liu F,Ng TB,Liu ZK
Mon Jul 31 00:00:00 UTC 2023
New fungal protein from Pleurotus ferulae lanzi induces AMPK-mediated autophagy and G1-phase cell cycle arrest in A549 lung cancer cells.
International journal of biological macromolecules
Liu MH,Liu F,Ng TB,Liu ZK
Mon Jul 31 00:00:00 UTC 2023
RPL11 promotes non-small cell lung cancer cell proliferation by regulating endoplasmic reticulum stress and cell autophagy.
BMC molecular and cell biology
Chen J,Lei C,Zhang H,Huang X,Yang Y,Liu J,Jia Y,Shi H,Zhang Y,Zhang J,Du J
Fri Mar 03 00:00:00 UTC 2023
Co-exposure to environmentally relevant concentrations of cadmium and polystyrene nanoplastics induced oxidative stress, ferroptosis and excessive mitophagy in mice kidney.
Environmental pollution (Barking, Essex : 1987)
Qiu W,Ye J,Su Y,Zhang X,Pang X,Liao J,Wang R,Zhao C,Zhang H,Hu L,Tang Z,Su R
Fri Sep 15 00:00:00 UTC 2023
Benzo[a]pyrene induces epithelial tight junction disruption and apoptosis via inhibiting the initiation of autophagy in intestinal porcine epithelial cells.
Chemico-biological interactions
Li J,Bai J,Si X,Jia H,Wu Z
Sat Apr 01 00:00:00 UTC 2023
hASCs-derived exosomal miR-155-5p targeting TGFβR2 promotes autophagy and reduces pyroptosis to alleviate intervertebral disc degeneration.
Journal of orthopaedic translation
Chen D,Jiang X,Zou H
Wed Mar 01 00:00:00 UTC 2023
Crosstalk between Mfn2-mediated mitochondria associated membranes disorder and autophagy induced by molybdenum and cadmium in sheep heart.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Peng C,Yang S,Yang F,Xiong Z,Liu Q,Liao S,Huang K,Wan W,Cao H
Sat Apr 01 00:00:00 UTC 2023
Acinetobacter baumannii outer membrane protein A induces autophagy in bone marrow-derived dendritic cells involving the PI3K/mTOR pathway.
Immunity, inflammation and disease
Tan H,Cao L
Sat Apr 01 00:00:00 UTC 2023
Corilagin alleviates intestinal ischemia/reperfusion injury by relieving oxidative stress and apoptosis via AMPK/Sirt1-autophagy pathway.
Experimental biology and medicine (Maywood, N.J.)
Li B,Li W,Zheng M,Wang Y,Diao Y,Mou X,Liu J
Wed Feb 01 00:00:00 UTC 2023
The cytotoxicity of zinc oxide nanoparticles to 3D brain organoids results from excessive intracellular zinc ions and defective autophagy.
Cell biology and toxicology
Liu L,Wang J,Zhang J,Huang C,Yang Z,Cao Y
Wed Feb 01 00:00:00 UTC 2023
Circadian gene CLOCK accelerates atherosclerosis by promoting endothelial autophagy.
Biotechnology & genetic engineering reviews
Wang C,Yang X,Guo Z,Zhu G,Fan L
Wed Mar 22 00:00:00 UTC 2023
Unlocking the potential of iridium and ruthenium arene complexes as anti-tumor and anti-metastasis chemotherapeutic agents.
Journal of inorganic biochemistry
Lv M,Qian X,Li S,Gong J,Wang Q,Qian Y,Su Z,Xue X,Liu HK
Sun Jan 01 00:00:00 UTC 2023
Bioengineered Nanospores Selectively Blocking LC3-Associated Phagocytosis in Tumor-Associated Macrophages Potentiate Antitumor Immunity.
ACS nano
Shang Y,Lu H,Liao L,Li S,Xiong H,Yao J
Tue Jun 13 00:00:00 UTC 2023
Role of nuclear factor of activated T Cells-1 in Sepsis-induced behavioral deficits in mice.
Brain research
Jiang F,Guo Y,Hu L,Zhang M,Meng J,Si Y,Shi H
Mon May 01 00:00:00 UTC 2023
Development of Clioquinol Platinum(IV) Conjugates as Autophagy-Targeted Antimetastatic Agents.
Journal of medicinal chemistry
Zhang M,Li L,Li S,Liu Z,Zhang N,Sun B,Wang Z,Jia D,Liu M,Wang Q
Thu Mar 09 00:00:00 UTC 2023
Jiedu Tongluo Baoshen formula enhances renal tubular epithelial cell autophagy to prevent renal fibrosis by activating SIRT1/LKB1/AMPK pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Jin D,Zhao Y,Sun Y,Xue J,Li X,Wang X
Sat Apr 01 00:00:00 UTC 2023
Abamectin causes toxicity to the carp respiratory system by triggering oxidative stress, inflammation, and apoptosis and inhibiting autophagy.
Environmental science and pollution research international
Feng H,Zhou P,Liu F,Zhang W,Yang H,Li X,Dong J
Sat Apr 01 00:00:00 UTC 2023
Marein reduces lipid levels via modulating the PI3K/AKT/mTOR pathway to induce lipophagy.
Journal of ethnopharmacology
Zhang PP,Zhuo BY,Duan ZW,Li X,Huang SL,Cao Q,Zhao T,Wei SL,Hu XH,Zhang Y
Thu Aug 10 00:00:00 UTC 2023
Strontium ion attenuates osteoarthritis through inhibiting senescence and enhancing autophagy in fibroblast-like synoviocytes.
Molecular biology reports
Liao B,Ding M,Wang Y,Xu H,Shangguan L
Wed Feb 01 00:00:00 UTC 2023
XBP1s gene of endoplasmic reticulum stress enhances proliferation and osteogenesis of human periodontal ligament cells.
Tissue & cell
Cui Z,Qin R,Feng J,Liu Y,Zhou X,Qin X,Li Y,Zhang Z,He X
Tue Aug 01 00:00:00 UTC 2023
TRIP13 overexpression promotes gefitinib resistance in non‑small cell lung cancer via regulating autophagy and phosphorylation of the EGFR signaling pathway.
Oncology reports
Xiao Z,Li M,Zhang X,Rong X,Xu H
Mon May 01 00:00:00 UTC 2023
TRIP13 overexpression promotes gefitinib resistance in non‑small cell lung cancer via regulating autophagy and phosphorylation of the EGFR signaling pathway.
Oncology reports
Xiao Z,Li M,Zhang X,Rong X,Xu H
Mon May 01 00:00:00 UTC 2023
Metformin Collaborates with PINK1/Mfn2 Overexpression to Prevent Cardiac Injury by Improving Mitochondrial Function.
Biology
Ma Z,Liu Z,Li X,Zhang H,Han D,Xiong W,Zhou H,Yang X,Zeng Q,Ren H,Xu D
Tue Apr 11 00:00:00 UTC 2023
Scalp acupuncture alleviates cerebral ischemic stroke-induced motor dysfunction in rats via regulating endoplasmic reticulum stress and ER-phagy.
Scientific reports
Zhang Y,Lou H,Lu J,Tang X,Pang T,Lei S,Cong D,Wang Y,Sun L
Wed Jun 21 00:00:00 UTC 2023
Anticancer efficacy of hirsuteine against colorectal cancer by opposite modulation of wild-type and mutant p53.
Discover. Oncology
Zhang Y,Guo T,Li S,Ren Z,Gao S,Lu H,Ma X,Liu D,Liu Y,Kong D,Qiu Y
Wed May 31 00:00:00 UTC 2023
Dual role of ANGPTL8 in promoting tumor cell proliferation and immune escape during hepatocarcinogenesis.
Oncogenesis
Gao Y,Yuan Y,Wen S,Chen Y,Zhang Z,Feng Y,Jiang B,Ma S,Hu R,Fang C,Ruan X,Yuan Y,Fang X,Luo C,Meng Z,Wang X,Guo X
Mon May 15 00:00:00 UTC 2023
Bavachin protects against diet-induced hepatic steatosis and obesity in mice.
Acta pharmacologica Sinica
Wei X,Lin L,Yuan QQ,Wang XY,Zhang Q,Zhang XM,Tang KC,Guo MY,Dong TY,Han W,Huang DK,Qi YL,Zhang M,Zhang HB
Sat Jul 01 00:00:00 UTC 2023
Urolithin A reduces amyloid-beta load and improves cognitive deficits uncorrelated with plaque burden in a mouse model of Alzheimer's disease.
GeroScience
Ballesteros-Álvarez J,Nguyen W,Sivapatham R,Rane A,Andersen JK
Sat Apr 01 00:00:00 UTC 2023
MALAT1-miR-30c-5p-CTGF/ATG5 axis regulates silica-induced experimental silicosis by mediating EMT in alveolar epithelial cells.
Ecotoxicology and environmental safety
Xia J,Tian Y,Shao Z,Li C,Ding M,Qi Y,Xu X,Dai K,Wu C,Yao W,Hao C
Sun Jan 01 00:00:00 UTC 2023
MALAT1-miR-30c-5p-CTGF/ATG5 axis regulates silica-induced experimental silicosis by mediating EMT in alveolar epithelial cells.
Ecotoxicology and environmental safety
Xia J,Tian Y,Shao Z,Li C,Ding M,Qi Y,Xu X,Dai K,Wu C,Yao W,Hao C
Sun Jan 01 00:00:00 UTC 2023
The COPS3-FOXO3 positive feedback loop regulates autophagy to promote cisplatin resistance in osteosarcoma.
Autophagy
Niu J,Yan T,Guo W,Wang W,Ren T,Huang Y,Zhao Z,Yu Y,Chen C,Huang Q,Lou J,Guo L
Thu Jun 01 00:00:00 UTC 2023
The COPS3-FOXO3 positive feedback loop regulates autophagy to promote cisplatin resistance in osteosarcoma.
Autophagy
Niu J,Yan T,Guo W,Wang W,Ren T,Huang Y,Zhao Z,Yu Y,Chen C,Huang Q,Lou J,Guo L
Thu Jun 01 00:00:00 UTC 2023
The COPS3-FOXO3 positive feedback loop regulates autophagy to promote cisplatin resistance in osteosarcoma.
Autophagy
Niu J,Yan T,Guo W,Wang W,Ren T,Huang Y,Zhao Z,Yu Y,Chen C,Huang Q,Lou J,Guo L
Thu Jun 01 00:00:00 UTC 2023
Time-dependent effect of REV-ERBα agonist SR9009 on nonalcoholic steatohepatitis and gut microbiota in mice.
Chronobiology international
Ni Y,Nan S,Zheng L,Zhang L,Zhao Y,Fu Z
Sat Jun 03 00:00:00 UTC 2023
Impaired autophagy flux by lncRNA NEAT1 is critical for inflammation factors production in human periodontal ligament stem cells with nicotine treatment.
Journal of periodontal research
Zhang T,Yang K,Chen Y,Jiang Y,Zhou Z,Liu J,Du Y,Wang L,Han X,Wu X,Wang X
Wed Feb 01 00:00:00 UTC 2023
Impaired autophagy flux by lncRNA NEAT1 is critical for inflammation factors production in human periodontal ligament stem cells with nicotine treatment.
Journal of periodontal research
Zhang T,Yang K,Chen Y,Jiang Y,Zhou Z,Liu J,Du Y,Wang L,Han X,Wu X,Wang X
Wed Feb 01 00:00:00 UTC 2023
Marine-Derived Natural Product HDYL-GQQ-495 Targets P62 to Inhibit Autophagy.
Marine drugs
Li Q,Fan J,Chen Y,Liu Y,Liu H,Jiang W,Li D,Dang Y
Fri Jan 20 00:00:00 UTC 2023
Exogenous H2S Attenuates Hypertension by Regulating Renin Exocytosis under Hyperglycaemic and Hyperlipidaemic Conditions.
International journal of molecular sciences
Liu N,Li M,Liu S,Kang J,Chen L,Huang J,Wang Y,Chen H,Zhang W
Sat Jan 14 00:00:00 UTC 2023
m6A reader HNRNPA2B1 destabilization of ATG4B regulates autophagic activity, proliferation and olaparib sensitivity in breast cancer.
Experimental cell research
Zheng R,Yu Y,Lv L,Zhang Y,Deng H,Li J,Zhang B
Wed Mar 01 00:00:00 UTC 2023
Natural Oncolysis of Enterovirus 71 in Antitumor Therapy of Colorectal Cancer.
Advanced biology
Ruan Z,Chi J,Kong Y,Li C,Ruan X,Zhou X,Chen Y,Li Y,Luo Z
Sat Jul 01 00:00:00 UTC 2023
Immunostimulation with chemotherapy of a ruthenium-arene complex via blockading CD47 signal in chronic myelogenous leukemia cells.
Journal of inorganic biochemistry
Hu G,Lv M,Guo B,Huang Y,Su Z,Qian Y,Xue X,Liu HK
Thu Jun 01 00:00:00 UTC 2023
SIRT6 drives sensitivity to ferroptosis in anaplastic thyroid cancer through NCOA4-dependent autophagy.
American journal of cancer research
Yang Z,Huang R,Wang Y,Guan Q,Li D,Wu Y,Liao T,Wang Y,Xiang J
Sat Mar 11 00:00:00 UTC 2023
Protective Effect and Mechanism of Kuntai Capsule on Angiotensin II -Induced Hypertension in Ovariectomized Rats.
Chinese journal of integrative medicine
Ge XF,Li SS,Liu YH,Lu MQ,Su HN,Yang X,Sun XW
Thu Jun 01 00:00:00 UTC 2023
Ginsenoside Rg1 alleviates learning and memory impairments and Aβ disposition through inhibiting NLRP1 inflammasome and autophagy dysfunction in APP/PS1 mice.
Molecular medicine reports
Li X,Huang L,Kong L,Su Y,Zhou H,Ji P,Sun R,Wang C,Li W,Li W
Sun Jan 01 00:00:00 UTC 2023
Ginsenoside Rg1 alleviates learning and memory impairments and Aβ disposition through inhibiting NLRP1 inflammasome and autophagy dysfunction in APP/PS1 mice.
Molecular medicine reports
Li X,Huang L,Kong L,Su Y,Zhou H,Ji P,Sun R,Wang C,Li W,Li W
Sun Jan 01 00:00:00 UTC 2023
Metformin regulates autophagy via LGMN to inhibit choriocarcinoma.
Gene
Tu W,Qin M,Li Y,Wu W,Tong X
Wed Feb 15 00:00:00 UTC 2023
Ambra1 modulates the sensitivity of mantle cell lymphoma to palbociclib by regulating cyclin D1.
Scientific reports
Jiang Z,Zhang A,Wei W,Li S
Wed May 24 00:00:00 UTC 2023
Ambra1 modulates the sensitivity of mantle cell lymphoma to palbociclib by regulating cyclin D1.
Scientific reports
Jiang Z,Zhang A,Wei W,Li S
Wed May 24 00:00:00 UTC 2023
Inhibition of PI3K/Akt/mTOR signaling by NDRG2 contributes to neuronal apoptosis and autophagy in ischemic stroke.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association
Wang Y,Wang B,Liu Y,Guo Y,Lu H,Liu X
Wed Mar 01 00:00:00 UTC 2023
A Novel Tri-Functional Liposome Re-Educates "Cold Tumor" and Abrogates Tumor Growth by Synergizing Autophagy Inhibition and PD-L1 Blockade.
Advanced healthcare materials
Zhou F,Li X,Xue X,Li S,Fan G,Cai Y,Chang Z,Qu J,Liu R
Sat Apr 01 00:00:00 UTC 2023
DR1-CSE/H2S pathway upregulates autophagy and inhibits H9C2 cells damage induced by high glucose.
Acta cardiologica
Li H,Wei Y,Xi Y,Jiao L,Wen X,Wu R,Chang G,Sun F,Hao J
Sat Jul 01 00:00:00 UTC 2023
Hypothermic Oxygenated Machine Perfusion Promotes Mitophagy Flux against Hypoxia-Ischemic Injury in Rat DCD Liver.
International journal of molecular sciences
Luo J,Hu Y,Qiao Y,Li H,Huang J,Xu K,Jiang L,Wu H,Hu X,Jia J,Zhou L,Xie H,Li J,Zheng S
Sat Mar 11 00:00:00 UTC 2023
Ginsenoside Rb1 induces autophagic lipid degradation via miR-128 targeting TFEB.
Food & function
Meng Z,Lu J,Ge G,Wang G,Zhang R,Li Y,Guan S,Lu J
Tue Jan 03 00:00:00 UTC 2023
Ginsenoside Rb1 induces autophagic lipid degradation via miR-128 targeting TFEB.
Food & function
Meng Z,Lu J,Ge G,Wang G,Zhang R,Li Y,Guan S,Lu J
Tue Jan 03 00:00:00 UTC 2023
Ginsenoside Rb1 induces autophagic lipid degradation via miR-128 targeting TFEB.
Food & function
Meng Z,Lu J,Ge G,Wang G,Zhang R,Li Y,Guan S,Lu J
Tue Jan 03 00:00:00 UTC 2023
Ginsenoside Rb1 induces autophagic lipid degradation via miR-128 targeting TFEB.
Food & function
Meng Z,Lu J,Ge G,Wang G,Zhang R,Li Y,Guan S,Lu J
Tue Jan 03 00:00:00 UTC 2023
Long-term copper exposure caused hepatocytes autophagy in broiler via miR-455-3p-OXSR1 axis.
Chemico-biological interactions
Lei C,Huo Y,Ma F,Liao J,Hu Z,Han Q,Li Y,Pan J,Hu L,Guo J,Tang Z
Thu Jan 05 00:00:00 UTC 2023
PRKAA1 induces aberrant mitophagy in a PINK1/Parkin-dependent manner, contributing to fluoride-induced developmental neurotoxicity.
Ecotoxicology and environmental safety
Tang Y,Zhang J,Hu Z,Xu W,Xu P,Ma Y,Xing H,Niu Q
Sat Apr 15 00:00:00 UTC 2023
A small molecule inhibitor of Notch1 modulates stemness and suppresses breast cancer cell growth.
Frontiers in pharmacology
Saran U,Chandrasekaran B,Tyagi A,Shukla V,Singh A,Sharma AK,Damodaran C
Fri May 19 00:00:00 UTC 2023
Neuro death through autophagy via the acetylation of FoxO1 by SIRT2 in the hippocampus of mice in a autism spectrum disorder mice model.
Journal of cellular physiology
Guo L,Jiang ZM,Zhan YJ,Pan W,Wu QW,Song FX,Zhou X,Zhou XY,Liu LJ,Wang JT
Thu Jun 01 00:00:00 UTC 2023
A high-concentrate diet induces inflammatory injury via regulating Ca2+/CaMKKβ-mediated autophagy in mammary gland tissue of dairy cows.
Frontiers in immunology
Meng M,Li X,Wang Z,Huo R,Ma N,Chang G,Shen X
Fri May 19 00:00:00 UTC 2023
Pre-ischaemic Treatment with Enriched Environment Alleviates Acute Neuronal Injury by Inhibiting Endoplasmic Reticulum Stress-dependent Autophagy and Apoptosis.
Neuroscience
Han Y,Shen XY,Gao ZK,Ping Han P,Bi X
Wed Mar 01 00:00:00 UTC 2023
Pre-ischaemic Treatment with Enriched Environment Alleviates Acute Neuronal Injury by Inhibiting Endoplasmic Reticulum Stress-dependent Autophagy and Apoptosis.
Neuroscience
Han Y,Shen XY,Gao ZK,Ping Han P,Bi X
Wed Mar 01 00:00:00 UTC 2023
TFEB coordinates autophagy and pyroptosis as hepatotoxicity responses to ZnO nanoparticles.
The Science of the total environment
Pei X,Liu D,Li J,Li L,Ding X,Zhang W,Li Z,Xu G,Li C,Li D
Mon Mar 20 00:00:00 UTC 2023
Long-term effect of PBDE-99 prenatal exposure on spermatogenic injuries via the dysregulation of autophagy.
Journal of hazardous materials
Wu J,Deng F,Tang X,Chen W,Zhou R,Zhao T,Mao X,Shu F
Thu Jun 15 00:00:00 UTC 2023
Long-term effect of PBDE-99 prenatal exposure on spermatogenic injuries via the dysregulation of autophagy.
Journal of hazardous materials
Wu J,Deng F,Tang X,Chen W,Zhou R,Zhao T,Mao X,Shu F
Thu Jun 15 00:00:00 UTC 2023
The ZSWIM8 ubiquitin ligase regulates neurodevelopment by guarding the protein quality of intrinsically disordered Dab1.
Cerebral cortex (New York, N.Y. : 1991)
Wang G,Lei J,Wang Y,Yu J,He Y,Zhao W,Hu Z,Xu Z,Jin Y,Gu Y,Guo X,Yang B,Gao Z,Wang Z
Tue Mar 21 00:00:00 UTC 2023
Host Restriction Factor A3G Inhibits the Replication of Enterovirus D68 by Competitively Binding the 5' Untranslated Region with PCBP1.
Journal of virology
Li Z,Yang X,Zhao Z,Liu X,Zhang W
Wed Jan 26 00:00:00 UTC 2022
Oridonin relieves depressive-like behaviors by inhibiting neuroinflammation and autophagy impairment in rats subjected to chronic unpredictable mild stress.
Phytotherapy research : PTR
Liang L,Wang H,Hu Y,Bian H,Xiao L,Wang G
Mon Aug 01 00:00:00 UTC 2022
Ablation of Discoidin Domain Receptor 1 Provokes an Osteopenic Phenotype by Regulating Osteoblast/Osteocyte Autophagy and Apoptosis.
Biomedicines
Chou HC,Lin SY,Chou LY,Ho ML,Chuang SC,Cheng TL,Kang L,Lin YS,Wang YH,Wei CW,Chen CH,Wang CZ
Fri Sep 02 00:00:00 UTC 2022
CAV1 alleviated CaOx stones formation via suppressing autophagy-dependent ferroptosis.
PeerJ
Yang Y,Hong S,Lu Y,Wang Q,Wang S,Xun Y
Wed Jan 18 00:00:00 UTC 2023
LINC-PINT suppresses cisplatin resistance in gastric cancer by inhibiting autophagy activation via epigenetic silencing of ATG5 by EZH2.
Frontiers in pharmacology
Zhang C,Kang T,Wang X,Wang J,Liu L,Zhang J,Liu X,Li R,Wang J,Zhang J
Tue Sep 13 00:00:00 UTC 2022
Mycoplasma hyopneumoniae Infection Activates the NOD1 Signaling Pathway to Modulate Inflammation.
Frontiers in cellular and infection microbiology
Liu W,Jiang P,Yang K,Song Q,Yuan F,Liu Z,Gao T,Zhou D,Guo R,Li C,Sun P,Tian Y
Tue Jul 26 00:00:00 UTC 2022
TRPC1 contributes to endotoxemia-induced myocardial dysfunction via mediating myocardial apoptosis and autophagy.
Pharmacological research
Tian W,Liu SY,Zhang M,Meng JR,Tang N,Feng YD,Sun Y,Gao YY,Zhou L,Cao W,Li XQ
Fri Jul 01 00:00:00 UTC 2022
Quercus acutissima Carruth. root extract triggers apoptosis, autophagy and inhibits cell viability in breast cancer cells.
Journal of ethnopharmacology
Ding W,Chen C,Li J,Geng X,Zhang H,Sun Y
Tue May 10 00:00:00 UTC 2022
Difenoconazole disrupts the blood-brain barrier and results in neurotoxicity in carp by inhibiting the Nrf2 pathway mediated ROS accumulation.
Ecotoxicology and environmental safety
Liu F,Wang Y,Chen L,Bello BK,Zhang T,Yang H,Li X,Pan E,Feng H,Dong J
Sat Oct 01 00:00:00 UTC 2022
Autism spectrum disorder-like behavior induced in rat offspring by perinatal exposure to di-(2-ethylhexyl) phthalate.
Environmental science and pollution research international
Li Y,Zhao Y,Lu Y,Lu X,Hu Y,Li Q,Shuai M,Li R
Fri Jul 01 00:00:00 UTC 2022
Domain fusion TLR2-4 enhances the autophagy-dependent clearance of Staphylococcus aureus in the genetic engineering goat.
eLife
Wang M,Qi Y,Cao Y,Zhang X,Wang Y,Liu Q,Zhang J,Zhou G,Ai Y,Wei S,Wang L,Liu G,Lian Z,Han H
Tue Jun 28 00:00:00 UTC 2022
Tp47 promoted the phagocytosis of HMC3 cells though autophagy induced by endoplamic reticlum stress.
Journal of the European Academy of Dermatology and Venereology : JEADV
Li W,Li QL,Xu QY,Wang XT,Yang TC
Tue Nov 01 00:00:00 UTC 2022
Inhibition of CDK1 attenuates neuronal apoptosis and autophagy and confers neuroprotection after chronic spinal cord injury in vivo.
Journal of chemical neuroanatomy
Nie BX,Zhao G,Yuan XF,Yu LX,Zhang J,Yuan Y,Liu Y,Hu J,Song E,Zhou YC,Shu J
Sat Jan 01 00:00:00 UTC 2022
Inhibition of CDK1 attenuates neuronal apoptosis and autophagy and confers neuroprotection after chronic spinal cord injury in vivo.
Journal of chemical neuroanatomy
Nie BX,Zhao G,Yuan XF,Yu LX,Zhang J,Yuan Y,Liu Y,Hu J,Song E,Zhou YC,Shu J
Sat Jan 01 00:00:00 UTC 2022
NUPR1 contributes to radiation resistance by maintaining ROS homeostasis via AhR/CYP signal axis in hepatocellular carcinoma.
BMC medicine
Zhan Y,Zhang Z,Liu Y,Fang Y,Xie Y,Zheng Y,Li G,Liang L,Ding Y
Wed Oct 19 00:00:00 UTC 2022
FAT4 activation inhibits epithelial-mesenchymal transition (EMT) by promoting autophagy in H2228/Cer cells.
Medical oncology (Northwood, London, England)
Yang Y,Li Y,Yang Q,Liu Z,Chang X,Yang H,Liu J,Li Z,Zuo D
Wed Dec 28 00:00:00 UTC 2022
Beclin1 Deficiency Suppresses Epileptic Seizures.
Frontiers in molecular neuroscience
Yang M,Lin P,Jing W,Guo H,Chen H,Chen Y,Guo Y,Gu Y,He M,Wu J,Jiang X,Zou Z,Xu X,Chen C,Xiao F,Wang X,Tian X
Tue Aug 09 00:00:00 UTC 2022
1,3-Dichloro-2-propanol-Induced Renal Tubular Cell Necroptosis through the ROS/RIPK3/MLKL Pathway.
Journal of agricultural and food chemistry
Fan Y,Lu J,Yu Z,Qu X,Guan S
Wed Sep 07 00:00:00 UTC 2022
1,3-Dichloro-2-propanol-Induced Renal Tubular Cell Necroptosis through the ROS/RIPK3/MLKL Pathway.
Journal of agricultural and food chemistry
Fan Y,Lu J,Yu Z,Qu X,Guan S
Wed Sep 07 00:00:00 UTC 2022
TARDBP Inhibits Porcine Epidemic Diarrhea Virus Replication through Degrading Viral Nucleocapsid Protein and Activating Type I Interferon Signaling.
Journal of virology
Dong S,Kong N,Zhang Y,Li Y,Sun D,Qin W,Zhai H,Zhai X,Yang X,Ye C,Ye M,Liu C,Yu L,Zheng H,Tong W,Yu H,Zhang W,Tong G,Shan T
Wed May 25 00:00:00 UTC 2022
EPA and DHA differentially coordinate the crosstalk between host and gut microbiota and block DSS-induced colitis in mice by a reinforced colonic mucus barrier.
Food & function
Fang J,Zhang Z,Cheng Y,Yang H,Zhang H,Xue Z,Lu S,Dong Y,Song C,Zhang X,Zhou Y
Wed Apr 20 00:00:00 UTC 2022
Phosphoinositide-3-kinase regulatory subunit 4 participates in the occurrence and development of amyotrophic lateral sclerosis by regulating autophagy.
Neural regeneration research
Liu Y,Wei CH,Li C,Chen WZ,Zhu Y,Xu RS
Fri Jul 01 00:00:00 UTC 2022
Nodosin Exerts an Anti-Colorectal Cancer Effect by Inhibiting Proliferation and Triggering Complex Cell Death in Vitro and in Vivo.
Frontiers in pharmacology
Fan H,Hao X,Gao Y,Yang J,Liu A,Su Y,Xia Y
Tue Aug 09 00:00:00 UTC 2022
Nodosin Exerts an Anti-Colorectal Cancer Effect by Inhibiting Proliferation and Triggering Complex Cell Death in Vitro and in Vivo.
Frontiers in pharmacology
Fan H,Hao X,Gao Y,Yang J,Liu A,Su Y,Xia Y
Tue Aug 09 00:00:00 UTC 2022
Nodosin Exerts an Anti-Colorectal Cancer Effect by Inhibiting Proliferation and Triggering Complex Cell Death in Vitro and in Vivo.
Frontiers in pharmacology
Fan H,Hao X,Gao Y,Yang J,Liu A,Su Y,Xia Y
Tue Aug 09 00:00:00 UTC 2022
Nodosin Exerts an Anti-Colorectal Cancer Effect by Inhibiting Proliferation and Triggering Complex Cell Death in Vitro and in Vivo.
Frontiers in pharmacology
Fan H,Hao X,Gao Y,Yang J,Liu A,Su Y,Xia Y
Tue Aug 09 00:00:00 UTC 2022
Puerarin prevents calcium oxalate crystal-induced renal epithelial cell autophagy by activating the SIRT1-mediated signaling pathway.
Urolithiasis
Jing GH,Liu YD,Liu JN,Jin YS,Yu SL,An RH
Sat Oct 01 00:00:00 UTC 2022
Integrated Profiles of Transcriptome and mRNA m6A Modification Reveal the Intestinal Cytotoxicity of Aflatoxin B1 on HCT116 Cells.
Genes
Wu Y,Bao W,Ren J,Li C,Chen M,Zhang D,Zhu A
Tue Dec 27 00:00:00 UTC 2022
CN-3 increases TMZ sensitivity and induces ROS-dependent apoptosis and autophagy in TMZ-resistance glioblastoma.
Journal of biochemical and molecular toxicology
Xue YY,Lu YY,Sun GQ,Fang F,Ji YQ,Tang HF,Qiu PC,Cheng G
Tue Mar 01 00:00:00 UTC 2022
Tourniquet-induced lower limb ischemia/reperfusion reduces mitochondrial function by decreasing mitochondrial biogenesis in acute kidney injury in mice.
Physiological reports
Packialakshmi B,Stewart IJ,Burmeister DM,Feng Y,McDaniel DP,Chung KK,Zhou X
Tue Feb 01 00:00:00 UTC 2022
Bilobalide Exerts Anti-Inflammatory Effects on Chondrocytes Through the AMPK/SIRT1/mTOR Pathway to Attenuate ACLT-Induced Post-Traumatic Osteoarthritis in Rats.
Frontiers in pharmacology
Ma T,Lv L,Yu Y,Jia L,Song X,Xu X,Li T,Sheng X,Wang H,Zhang J,Gao L
Wed Mar 16 00:00:00 UTC 2022
Type I collagen reduces lipid accumulation during adipogenesis of preadipocytes 3T3-L1 via the YAP-mTOR-autophagy axis.
Biochimica et biophysica acta. Molecular and cell biology of lipids
Gao Y,Ma K,Kang Y,Liu W,Liu X,Long X,Hayashi T,Hattori S,Mizuno K,Fujisaki H,Ikejima T
Thu Sep 01 00:00:00 UTC 2022
Downregulation of FAPP2 gene induces cell autophagy and inhibits PI3K/AKT/mTOR pathway in T-cell acute lymphoblastic leukemia.
Hematological oncology
Yuan T,Wang J,Shi C,Wang Y,Xia B,Xu W,Yang H,Yang Y,Ye MT,Khalid S,Liang Y,Tian C,You MJ,Wang Y
Fri Apr 01 00:00:00 UTC 2022
Downregulation of FAPP2 gene induces cell autophagy and inhibits PI3K/AKT/mTOR pathway in T-cell acute lymphoblastic leukemia.
Hematological oncology
Yuan T,Wang J,Shi C,Wang Y,Xia B,Xu W,Yang H,Yang Y,Ye MT,Khalid S,Liang Y,Tian C,You MJ,Wang Y
Fri Apr 01 00:00:00 UTC 2022
Inhibition of Ciliogenesis Enhances the Cellular Sensitivity to Temozolomide and Ionizing Radiation in Human Glioblastoma Cells.
Biomedical and environmental sciences : BES
Wei L,Ma W,Cai H,Peng SP,Tian HB,Wang JF,Gao L,He JP
Fri May 20 00:00:00 UTC 2022
CXCR4 inhibition attenuates calcium oxalate crystal deposition-induced renal fibrosis.
International immunopharmacology
Ye Z,Xia Y,Zhou X,Li B,Yu W,Ruan Y,Li H,Ning J,Chen L,Rao T,Cheng F
Wed Jun 01 00:00:00 UTC 2022
IGF-1 Signaling Regulates Mitochondrial Remodeling during Myogenic Differentiation.
Nutrients
Guan X,Yan Q,Wang D,Du G,Zhou J
Wed Mar 16 00:00:00 UTC 2022
All-In-One Biomimetic Nanoplatform Based on Hollow Polydopamine Nanoparticles for Synergistically Enhanced Radiotherapy of Colon Cancer.
Small (Weinheim an der Bergstrasse, Germany)
Gong L,Zhang Y,Zhao J,Zhang Y,Tu K,Jiao L,Xu Q,Zhang M,Han S
Fri Apr 01 00:00:00 UTC 2022
Sesamin Ameliorates Fine Particulate Matter (PM2.5)-Induced Lung Injury via Suppression of Apoptosis and Autophagy in Rats.
Journal of agricultural and food chemistry
Ren J,Li X,Zhu S,Yin B,Guo Z,Cui Q,Song J,Pei H,Ma Y
Wed Aug 03 00:00:00 UTC 2022
Sesamin Ameliorates Fine Particulate Matter (PM2.5)-Induced Lung Injury via Suppression of Apoptosis and Autophagy in Rats.
Journal of agricultural and food chemistry
Ren J,Li X,Zhu S,Yin B,Guo Z,Cui Q,Song J,Pei H,Ma Y
Wed Aug 03 00:00:00 UTC 2022
Channel HCN4 mutation R666Q associated with sporadic arrhythmia decreases channel electrophysiological function and increases protein degradation.
The Journal of biological chemistry
Wang H,Wu T,Huang Z,Huang J,Geng Z,Cui B,Yan Y,Zhang Y,Wang Y
Tue Nov 01 00:00:00 UTC 2022
Drp1 knockdown represses apoptosis of rat retinal endothelial cells by inhibiting mitophagy.
Acta histochemica
Wu H,Li G,Chen W,Luo W,Yang Z,You Z,Zou Y
Sat Jan 01 00:00:00 UTC 2022
Polyphyllin II induced apoptosis of NSCLC cells by inhibiting autophagy through the mTOR pathway.
Pharmaceutical biology
Jiao Y,Xin M,Xu J,Xiang X,Li X,Jiang J,Jia X
Thu Dec 01 00:00:00 UTC 2022
Trappc9 Deficiency Impairs the Plasticity of Stem Cells.
International journal of molecular sciences
Usman M,Li Y,Ke Y,Chhetri G,Islam MA,Wang Z,Li X
Thu Apr 28 00:00:00 UTC 2022
TFEB ameliorates autophagy flux disturbance induced by PBDE-47 via up-regulating autophagy-lysosome fusion.
Journal of hazardous materials
Tang H,Huang H,Wang D,Li P,Tian Z,Li D,Wang S,Ma R,Xia T,Wang A
Sun May 15 00:00:00 UTC 2022
TFEB ameliorates autophagy flux disturbance induced by PBDE-47 via up-regulating autophagy-lysosome fusion.
Journal of hazardous materials
Tang H,Huang H,Wang D,Li P,Tian Z,Li D,Wang S,Ma R,Xia T,Wang A
Sun May 15 00:00:00 UTC 2022
Gaseous signal molecule SO2 regulates autophagy through PI3K/AKT pathway inhibits cardiomyocyte apoptosis and improves myocardial fibrosis in rats with type II diabetes.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology
Zhao J,Wu Q,Yang T,Nie L,Liu S,Zhou J,Chen J,Jiang Z,Xiao T,Yang J,Chu C
Tue Nov 01 00:00:00 UTC 2022
HIF-1α-mediated autophagy and canonical Wnt/β-catenin signalling activation are involved in fluoride-induced osteosclerosis in rats.
Environmental pollution (Barking, Essex : 1987)
Zhu S,Liu J,Zhao J,Zhou B,Zhang Y,Wang H
Thu Dec 15 00:00:00 UTC 2022
FUBP3 Degrades the Porcine Epidemic Diarrhea Virus Nucleocapsid Protein and Induces the Production of Type I Interferon.
Journal of virology
Dong S,Kong N,Wang C,Li Y,Sun D,Qin W,Zhai H,Zhai X,Yang X,Ye C,Ye M,Liu C,Yu L,Zheng H,Tong W,Yu H,Zhang W,Tong G,Shan T
Wed Jul 13 00:00:00 UTC 2022
Curcumin exerts chondroprotective effects against osteoarthritis by promoting AMPK/PINK1/Parkin-mediated mitophagy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Jin Z,Chang B,Wei Y,Yang Y,Zhang H,Liu J,Piao L,Bai L
Fri Jul 01 00:00:00 UTC 2022
The autophagic protein p62 is a target of reactive aldehydes in human and murine cholestatic liver disease.
PloS one
Shearn CT,Anderson AL,Devereux MW,Orlicky DJ,Michel C,Petersen DR,Miller CG,Harpavat S,Schmidt EE,Sokol RJ
Fri Nov 18 00:00:00 UTC 2022
Interpreting the Pharmacological Mechanisms of Sho-saiko-to on Thyroid Carcinoma through Combining Network Pharmacology and Experimental Evaluation.
ACS omega
Wang K,Qian R,Li H,Wang C,Ding Y,Gao Z
Tue Apr 05 00:00:00 UTC 2022
Protein disulfide isomerase blocks the interaction of LC3II-PHB2 and promotes mTOR signaling to regulate autophagy and radio/chemo-sensitivity.
Cell death & disease
Wang R,Shang Y,Chen B,Xu F,Zhang J,Zhang Z,Zhao X,Wan X,Xu A,Wu L,Zhao G
Thu Oct 06 00:00:00 UTC 2022
2-Deoxy-D-glucose Alleviates Cancer Cachexia-Induced Muscle Wasting by Enhancing Ketone Metabolism and Inhibiting the Cori Cycle.
Cells
Wei L,Wang R,Wazir J,Lin K,Song S,Li L,Pu W,Zhao C,Wang Y,Su Z,Wang H
Sun Sep 25 00:00:00 UTC 2022
2-Deoxy-D-glucose Alleviates Cancer Cachexia-Induced Muscle Wasting by Enhancing Ketone Metabolism and Inhibiting the Cori Cycle.
Cells
Wei L,Wang R,Wazir J,Lin K,Song S,Li L,Pu W,Zhao C,Wang Y,Su Z,Wang H
Sun Sep 25 00:00:00 UTC 2022
MgIG exerts therapeutic effects on crizotinib-induced hepatotoxicity by limiting ROS-mediated autophagy and pyroptosis.
Journal of cellular and molecular medicine
Li M,Wang C,Yu Z,Lan Q,Xu S,Ye Z,Li R,Ying L,Zhang X,Zhou Z
Mon Aug 01 00:00:00 UTC 2022
Exercise Ameliorates Atherosclerosis via Up-Regulating Serum β-Hydroxybutyrate Levels.
International journal of molecular sciences
Xu Z,Zhang M,Li X,Wang Y,Du R
Wed Mar 30 00:00:00 UTC 2022
Exercise Ameliorates Atherosclerosis via Up-Regulating Serum β-Hydroxybutyrate Levels.
International journal of molecular sciences
Xu Z,Zhang M,Li X,Wang Y,Du R
Wed Mar 30 00:00:00 UTC 2022
Estrogen receptor β activation inhibits colitis by promoting NLRP6-mediated autophagy.
Cell reports
Fan W,Ding C,Liu S,Gao X,Shen X,De Boevre M,Gao Z,Li M,Zhang S,Miao Y,Guan W,Liu G,Yan L,De Saeger S,Song S
Tue Oct 11 00:00:00 UTC 2022
Estrogen receptor β activation inhibits colitis by promoting NLRP6-mediated autophagy.
Cell reports
Fan W,Ding C,Liu S,Gao X,Shen X,De Boevre M,Gao Z,Li M,Zhang S,Miao Y,Guan W,Liu G,Yan L,De Saeger S,Song S
Tue Oct 11 00:00:00 UTC 2022
Docosahexaenoic Acid-Enhanced Autophagic Flux Improves Cardiac Dysfunction after Myocardial Infarction by Targeting the AMPK/mTOR Signaling Pathway.
Oxidative medicine and cellular longevity
Shi Y,Li H,Wu T,Wang Q,Zhu Q,Guan X,Wu R
Thu Mar 24 00:00:00 UTC 2022
Huaier suppresses pancreatic cancer progression via activating cell autophagy induced ferroptosis.
Frontiers in oncology
Zhu Z,Wang X,Zhang W,Gong M,Zhang S,Yang B,Qu B,Wu Z,Ma Q,Wang Z,Qian W
Wed Oct 19 00:00:00 UTC 2022
Huaier suppresses pancreatic cancer progression via activating cell autophagy induced ferroptosis.
Frontiers in oncology
Zhu Z,Wang X,Zhang W,Gong M,Zhang S,Yang B,Qu B,Wu Z,Ma Q,Wang Z,Qian W
Wed Oct 19 00:00:00 UTC 2022
Huaier suppresses pancreatic cancer progression via activating cell autophagy induced ferroptosis.
Frontiers in oncology
Zhu Z,Wang X,Zhang W,Gong M,Zhang S,Yang B,Qu B,Wu Z,Ma Q,Wang Z,Qian W
Wed Oct 19 00:00:00 UTC 2022
FTO-dependent N(6)-Methyladenosine regulates the progression of endometriosis via the ATG5/PKM2 Axis.
Cellular signalling
Wang H,Liang Z,Gou Y,Li Z,Cao Y,Jiao N,Tan J,Yu Y,Zhang Z
Sat Oct 01 00:00:00 UTC 2022
Creatine modulates cellular energy metabolism and protects against cancer cachexia-associated muscle wasting.
Frontiers in pharmacology
Wei L,Wang R,Lin K,Jin X,Li L,Wazir J,Pu W,Lian P,Lu R,Song S,Zhao Q,Li J,Wang H
Tue Dec 27 00:00:00 UTC 2022
Creatine modulates cellular energy metabolism and protects against cancer cachexia-associated muscle wasting.
Frontiers in pharmacology
Wei L,Wang R,Lin K,Jin X,Li L,Wazir J,Pu W,Lian P,Lu R,Song S,Zhao Q,Li J,Wang H
Tue Dec 27 00:00:00 UTC 2022
Paeoniflorin binds to VEGFR2 to restore autophagy and inhibit apoptosis for podocyte protection in diabetic kidney disease through PI3K-AKT signaling pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang X,Jiang L,Liu XQ,Huang YB,Wang AL,Zeng HX,Gao L,Zhu QJ,Xia LL,Wu YG
Tue Nov 01 00:00:00 UTC 2022
Modification of taxifolin particles with an enteric coating material promotes repair of acute liver injury in mice through modulation of inflammation and autophagy signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Ding Q,Chen K,Liu X,Ding C,Zhao Y,Sun S,Zhang Y,Zhang J,Liu S,Liu W
Mon Aug 01 00:00:00 UTC 2022
Network Pharmacology Research and Dual-omic Analyses Reveal the Molecular Mechanism of Natural Product Nodosin Inhibiting Muscle-Invasive Bladder Cancer in Vitro and in Vivo.
Journal of natural products
Hao X,Fan H,Yang J,Tang J,Zhou J,Zhao Y,Huang L,Xia Y
Fri Aug 26 00:00:00 UTC 2022
Network Pharmacology Research and Dual-omic Analyses Reveal the Molecular Mechanism of Natural Product Nodosin Inhibiting Muscle-Invasive Bladder Cancer in Vitro and in Vivo.
Journal of natural products
Hao X,Fan H,Yang J,Tang J,Zhou J,Zhao Y,Huang L,Xia Y
Fri Aug 26 00:00:00 UTC 2022
Lycorine improves peripheral nerve function by promoting Schwann cell autophagy via AMPK pathway activation and MMP9 downregulation in diabetic peripheral neuropathy.
Pharmacological research
Yuan Q,Zhang X,Wei W,Zhao J,Wu Y,Zhao S,Zhu L,Wang P,Hao J
Sat Jan 01 00:00:00 UTC 2022
Lycorine improves peripheral nerve function by promoting Schwann cell autophagy via AMPK pathway activation and MMP9 downregulation in diabetic peripheral neuropathy.
Pharmacological research
Yuan Q,Zhang X,Wei W,Zhao J,Wu Y,Zhao S,Zhu L,Wang P,Hao J
Sat Jan 01 00:00:00 UTC 2022
Heidihuangwan alleviates renal fibrosis in rats with 5/6 nephrectomy by inhibiting autophagy.
Frontiers in pharmacology
Li YY,Tian ZH,Pan GH,Zhao P,Pan DJ,Zhang JQ,Ye LY,Zhang FR,Xu XD
Wed Sep 28 00:00:00 UTC 2022
Heidihuangwan alleviates renal fibrosis in rats with 5/6 nephrectomy by inhibiting autophagy.
Frontiers in pharmacology
Li YY,Tian ZH,Pan GH,Zhao P,Pan DJ,Zhang JQ,Ye LY,Zhang FR,Xu XD
Wed Sep 28 00:00:00 UTC 2022
Cellular communication network factor 1 interlinks autophagy and ERK signaling to promote osteogenesis of periodontal ligament stem cells.
Journal of periodontal research
Li Z,Wu Z,Xi X,Zhao F,Liu H,Liu D
Thu Dec 01 00:00:00 UTC 2022
Transcription factor EB enhances autophagy and ameliorates palmitate-induced insulin resistance at least partly via upregulating AMPK activity in skeletal muscle cells.
Clinical and experimental pharmacology & physiology
Wang P,Li CG,Zhou X,Ding S
Tue Feb 01 00:00:00 UTC 2022
Induction of estrogen receptor β-mediated autophagy sensitizes breast cancer cells to TAD1822-7, a novel biphenyl urea taspine derivative.
Molecular biology reports
Su Q,Wu Q,Chen K,Wang J,Sarwar A,Zhang Y
Tue Feb 01 00:00:00 UTC 2022
CHST7 Methylation Status Related to the Proliferation and Differentiation of Pituitary Adenomas.
Cells
Dong W,Shi W,Liu Y,Li J,Zhang Y,Dong G,Dong X,Gao H
Thu Aug 04 00:00:00 UTC 2022
Autophagy: A New Mechanism for Esketamine as a Depression Therapeutic.
Neuroscience
Jiang G,Wang Y,Liu Q,Gu T,Liu S,Yin A,Zhang L
Sun Aug 21 00:00:00 UTC 2022
Overexpression of LINC00551 promotes autophagy-dependent ferroptosis of lung adenocarcinoma via upregulating DDIT4 by sponging miR-4328.
PeerJ
Peng X,Yang R,Peng W,Zhao Z,Tu G,He B,Cai Q,Shi S,Yin W,Yu F,Tao Y,Wang X
Tue Dec 27 00:00:00 UTC 2022
Acetyl CoA synthase 2 potentiates ATG5-induced autophagy against neuronal apoptosis after subarachnoid hemorrhage.
Journal of molecular histology
He W,Zhou X,Wu Q,Zhou L,Zhang Z,Zhang R,Deng C,Zhang X
Fri Apr 01 00:00:00 UTC 2022
Amino acid starvation-induced LDLR trafficking accelerates lipoprotein endocytosis and LDL clearance.
EMBO reports
Chen Y,Wu X,Zhang J,Pan G,Wang X,Guo X,Wang J,Cui X,Gao H,Cheng M,Yang J,Zhang C,Jiang F
Thu Feb 03 00:00:00 UTC 2022
The inhibition of PGAM5 suppresses seizures in a kainate-induced epilepsy model via mitophagy reduction.
Frontiers in molecular neuroscience
Zhong F,Gan Y,Song J,Zhang W,Yuan S,Qin Z,Wu J,Lü Y,Yu W
Wed Jan 11 00:00:00 UTC 2023
Clinical, genetic, and pathological characterization of GNE myopathy in China.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology
Lv XQ,Xu L,Lin PF,Yan CZ
Fri Jul 01 00:00:00 UTC 2022
Elaiophylin triggers paraptosis and preferentially kills ovarian cancer drug-resistant cells by inducing MAPK hyperactivation.
Signal transduction and targeted therapy
Li GN,Zhao XJ,Wang Z,Luo MS,Shi SN,Yan DM,Li HY,Liu JH,Yang Y,Tan JH,Zhang ZY,Chen RQ,Lai HL,Huang XY,Zhou JF,Ma D,Fang Y,Gao QL
Mon Sep 12 00:00:00 UTC 2022
ROS function as an inducer of autophagy to promote granulocyte proliferation in Pacific oyster Crassostrea gigas.
Developmental and comparative immunology
Zhang D,Dong M,Song X,Qiao X,Yang Y,Yu S,Sun W,Wang L,Song L
Sat Oct 01 00:00:00 UTC 2022
The alleviative effect of flavonol-type Nrf2 activator rhamnazin from Physalis alkekengi L. var. franchetii (Mast.) Makino on pulmonary disorders.
Phytotherapy research : PTR
Wu XY,Wang T,Hu HX,Zhang K,Zhao Y,Zhao BB,Lou HX,Wang XN,Shen T
Fri Apr 01 00:00:00 UTC 2022
The alleviative effect of flavonol-type Nrf2 activator rhamnazin from Physalis alkekengi L. var. franchetii (Mast.) Makino on pulmonary disorders.
Phytotherapy research : PTR
Wu XY,Wang T,Hu HX,Zhang K,Zhao Y,Zhao BB,Lou HX,Wang XN,Shen T
Fri Apr 01 00:00:00 UTC 2022
Propofol inhibits the malignant development of osteosarcoma U2OS cells via AMPK/FΟΧO1-mediated autophagy.
Oncology letters
Dai L,Li S,Li X,Jiang B
Thu Sep 01 00:00:00 UTC 2022
Development of artesunate intelligent prodrug liposomes based on mitochondrial targeting strategy.
Journal of nanobiotechnology
Gu L,Zhang J,Liu D,Chen J,Liu S,Peng Q,Tian Y,Du M,Zhang J,Xiao W,Shen S,Wang J
Sat Aug 13 00:00:00 UTC 2022
Development of artesunate intelligent prodrug liposomes based on mitochondrial targeting strategy.
Journal of nanobiotechnology
Gu L,Zhang J,Liu D,Chen J,Liu S,Peng Q,Tian Y,Du M,Zhang J,Xiao W,Shen S,Wang J
Sat Aug 13 00:00:00 UTC 2022
Development of artesunate intelligent prodrug liposomes based on mitochondrial targeting strategy.
Journal of nanobiotechnology
Gu L,Zhang J,Liu D,Chen J,Liu S,Peng Q,Tian Y,Du M,Zhang J,Xiao W,Shen S,Wang J
Sat Aug 13 00:00:00 UTC 2022
Development of artesunate intelligent prodrug liposomes based on mitochondrial targeting strategy.
Journal of nanobiotechnology
Gu L,Zhang J,Liu D,Chen J,Liu S,Peng Q,Tian Y,Du M,Zhang J,Xiao W,Shen S,Wang J
Sat Aug 13 00:00:00 UTC 2022
Metformin carbon nanodots promote odontoblastic differentiation of dental pulp stem cells by pathway of autophagy.
Frontiers in bioengineering and biotechnology
Lu J,Li R,Ni S,Xie Y,Liu X,Zhang K,Li Y
Wed Sep 28 00:00:00 UTC 2022
CircRNF144B/miR-342-3p/FBXL11 axis reduced autophagy and promoted the progression of ovarian cancer by increasing the ubiquitination of Beclin-1.
Cell death & disease
Song W,Zeng Z,Zhang Y,Li H,Cheng H,Wang J,Wu F
Sat Oct 08 00:00:00 UTC 2022
CircRNF144B/miR-342-3p/FBXL11 axis reduced autophagy and promoted the progression of ovarian cancer by increasing the ubiquitination of Beclin-1.
Cell death & disease
Song W,Zeng Z,Zhang Y,Li H,Cheng H,Wang J,Wu F
Sat Oct 08 00:00:00 UTC 2022
BNIP3 mediates the different adaptive responses of fibroblast-like synovial cells to hypoxia in patients with osteoarthritis and rheumatoid arthritis.
Molecular medicine (Cambridge, Mass.)
Deng R,Wang Y,Bu Y,Wu H
Sat Jun 11 00:00:00 UTC 2022
Metformin alleviates bone loss in ovariectomized mice through inhibition of autophagy of osteoclast precursors mediated by E2F1.
Cell communication and signaling : CCS
Xie X,Hu L,Mi B,Xue H,Hu Y,Panayi AC,Endo Y,Chen L,Yan C,Lin Z,Li H,Zhou W,Liu G
Tue Oct 25 00:00:00 UTC 2022
Metformin alleviates bone loss in ovariectomized mice through inhibition of autophagy of osteoclast precursors mediated by E2F1.
Cell communication and signaling : CCS
Xie X,Hu L,Mi B,Xue H,Hu Y,Panayi AC,Endo Y,Chen L,Yan C,Lin Z,Li H,Zhou W,Liu G
Tue Oct 25 00:00:00 UTC 2022
Iron deficiency exacerbates aortic medial degeneration by inducing excessive mitochondrial fission.
Food & function
Zhong X,Wu Q,Wang Z,Zhang M,Zheng S,Shi F,Chen Y,Che Y,Yuan S,Xing K
Mon Jul 18 00:00:00 UTC 2022
Ferulic acid ameliorates acetaminophen-induced acute liver injury by promoting AMPK-mediated protective autophagy.
IUBMB life
Wu J,Zhou F,Fan G,Liu J,Wang Y,Xue X,Lyu X,Lin S,Li X
Thu Sep 01 00:00:00 UTC 2022
Effects of bicyclol on hepatic sinusoidal obstruction syndrome induced by Gynura segetum.
Journal of clinical laboratory analysis
Yao J,Wu J,Jia S,Shao J,Zhang X,Xu Z,Zhang H,Li H,Yao X
Thu Dec 01 00:00:00 UTC 2022
Modification of lysine deacetylation regulates curcumol-induced necroptosis through autophagy in hepatic stellate cells.
Phytotherapy research : PTR
Sun S,Li Z,Huan S,Kai J,Xia S,Su Y,Ji S,Chen A,Wang S,Xu X,Shao J,Zhang F,Zhang B,Zhang Z,Zheng S
Wed Jun 01 00:00:00 UTC 2022
Maternal obesity induces liver lipid accumulation of offspring through the lncRNA Lockd/mTOR autophagy pathway.
Molecular genetics and genomics : MGG
Chen R,Yang H,Song Y,Yu H,Zhang M,Rao W,Wang Y,Xiao X,Chen Q,He Q
Thu Sep 01 00:00:00 UTC 2022
sSTEAP4 regulates cellular homeostasis and improves high-fat-diet-caused oxidative stress in hepatocytes.
Life sciences
Zhu L,Li B,Chen D,Chen N,Xu L,Li Q,Chen X
Sun May 01 00:00:00 UTC 2022
Impaired V-ATPase leads to increased lysosomal pH, results in disrupted lysosomal degradation and autophagic flux blockage, contributes to fluoride-induced developmental neurotoxicity.
Ecotoxicology and environmental safety
Han X,Tang Y,Zhang Y,Zhang J,Hu Z,Xu W,Xu S,Niu Q
Sun May 01 00:00:00 UTC 2022
Rhodiola activates macrophage migration inhibitory factor to alleviate non-alcoholic fatty liver disease.
Life sciences
Liu J,Li D,Dun Y,Li H,Ripley-Gonzalez JW,Zhang J,Qiu L,You B,Liu S
Tue Nov 01 00:00:00 UTC 2022
RNA sequence analysis reveals pathways and candidate genes associated with pancreatic acinar cells injury in a mouse pancreatitis model.
Tissue & cell
Chaoqun H,Rong Y,Yunpeng P,Xiaole Z,Wanli G,Chenyuan S,Dongya H,Yi M,Qiang L
Thu Dec 01 00:00:00 UTC 2022
San-Huang-Yi-Shen Capsule Ameliorates Diabetic Kidney Disease through Inducing PINK1/Parkin-Mediated Mitophagy and Inhibiting the Activation of NLRP3 Signaling Pathway.
Journal of diabetes research
Li H,Wang Y,Su X,Wang Q,Zhang S,Sun W,Zhang T,Dong M,Zhang Z,Lv S
Mon Nov 28 00:00:00 UTC 2022
San-Huang-Yi-Shen Capsule Ameliorates Diabetic Kidney Disease through Inducing PINK1/Parkin-Mediated Mitophagy and Inhibiting the Activation of NLRP3 Signaling Pathway.
Journal of diabetes research
Li H,Wang Y,Su X,Wang Q,Zhang S,Sun W,Zhang T,Dong M,Zhang Z,Lv S
Mon Nov 28 00:00:00 UTC 2022
Dihydromyricetin protects against Doxorubicin-induced cardiotoxicity through activation of AMPK/mTOR pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li X,Wang X,Wang B,Chi W,Li Z,Zhang M,Shen Y,Liu X,Lu Y,Liu Y
Sun May 01 00:00:00 UTC 2022
Dihydromyricetin protects against Doxorubicin-induced cardiotoxicity through activation of AMPK/mTOR pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li X,Wang X,Wang B,Chi W,Li Z,Zhang M,Shen Y,Liu X,Lu Y,Liu Y
Sun May 01 00:00:00 UTC 2022
Imiquimod induced vitiligo-like lesions-A consequence of modified melanocyte function.
Immunity, inflammation and disease
Yu H,Cen J,Lin X,Cheng H,Seifert O
Sat Jan 01 00:00:00 UTC 2022
CCL20/CCR6 axis mediates macrophages to promote proliferation and migration of ESCs by blocking autophagic flux in endometriosis.
Stem cell research & therapy
Tan J,Xu T,Gou Y,Wang H,Liang Z,Cao Y,Wang H,Yu Y,Jiao N,Zhang Z
Fri Jul 15 00:00:00 UTC 2022
CCL20/CCR6 axis mediates macrophages to promote proliferation and migration of ESCs by blocking autophagic flux in endometriosis.
Stem cell research & therapy
Tan J,Xu T,Gou Y,Wang H,Liang Z,Cao Y,Wang H,Yu Y,Jiao N,Zhang Z
Fri Jul 15 00:00:00 UTC 2022
Autophagy Promotes α-Amanitin-Induced Apoptosis of Hepa1-6 Liver Cells.
Chemical research in toxicology
Gu X,Zhang L,Sun W,Liu K,Xu H,Wu P,Gui M,Qu W
Mon Mar 21 00:00:00 UTC 2022
T-2 Toxin Induces Apoptotic Cell Death and Protective Autophagy in Mouse Microglia BV2 Cells.
Journal of fungi (Basel, Switzerland)
Sun T,Zhang Q,Li M,Tang S,Dai C
Fri Jul 22 00:00:00 UTC 2022
Gga-miR-30c-5p Suppresses Avian Reovirus (ARV) Replication by Inhibition of ARV-Induced Autophagy via Targeting ATG5.
Journal of virology
Zhou L,Haiyilati A,Li J,Li X,Gao L,Cao H,Wang Y,Zheng SJ
Wed Jul 27 00:00:00 UTC 2022
Prunella vulgaris can improve the pregnancy outcomes of experimental autoimmune thyroiditis rats by inhibiting Th1/Th17 immune responses.
Journal of reproductive immunology
Zhu Q,Muyayalo KP,Xu QH,Wang J,Wang H,Liao AH
Tue Feb 01 00:00:00 UTC 2022
Sulforaphane Ameliorates Limb Ischemia/Reperfusion-Induced Muscular Injury in Mice by Inhibiting Pyroptosis and Autophagy via the Nrf2-ARE Pathway.
Evidence-based complementary and alternative medicine : eCAM
Sun H,Wang J,Bi W,Zhang F,Chi K,Shi L,Yuan T,Ma K,Gao X
Thu Dec 14 00:00:00 UTC 2023
Let-7i-5p promotes a malignant phenotype in nasopharyngeal carcinoma via inhibiting tumor-suppressive autophagy.
Cancer letters
You B,Zhang P,Gu M,Yin H,Fan Y,Yao H,Pan S,Xie H,Cheng T,Liu H,You Y,Liu J
Sun Apr 10 00:00:00 UTC 2022
Let-7i-5p promotes a malignant phenotype in nasopharyngeal carcinoma via inhibiting tumor-suppressive autophagy.
Cancer letters
You B,Zhang P,Gu M,Yin H,Fan Y,Yao H,Pan S,Xie H,Cheng T,Liu H,You Y,Liu J
Sun Apr 10 00:00:00 UTC 2022
Let-7i-5p promotes a malignant phenotype in nasopharyngeal carcinoma via inhibiting tumor-suppressive autophagy.
Cancer letters
You B,Zhang P,Gu M,Yin H,Fan Y,Yao H,Pan S,Xie H,Cheng T,Liu H,You Y,Liu J
Sun Apr 10 00:00:00 UTC 2022
Trpc5-regulated AMPKα/mTOR autophagy pathway is associated with glucose metabolism disorders in low birth weight mice under overnutrition.
Biochemical and biophysical research communications
Zhou F,Yang L,Yang L,Wang X,Guo N,Sun W,Ma H
Sat Nov 19 00:00:00 UTC 2022
Apigenin alleviated PA-induced pyroptosis by activating autophagy in hepatocytes.
Food & function
Meng Z,Zhu B,Gao M,Wang G,Zhou H,Lu J,Guan S
Mon May 23 00:00:00 UTC 2022
Circ-PKD2 promotes Atg13-mediated autophagy by inhibiting miR-646 to increase the sensitivity of cisplatin in oral squamous cell carcinomas.
Cell death & disease
Gao L,Zhang Q,Li S,Zheng J,Ren W,Zhi K
Sat Feb 26 00:00:00 UTC 2022
Prototheca bovis induces autophagy in bovine mammary epithelial cells via the HIF-1α and AMPKα/ULK1 pathway.
Frontiers in immunology
Zhao W,Xu M,Barkema HW,Xie X,Lin Y,Khan S,Kastelic JP,Wang D,Deng Z,Han B
Mon Sep 26 00:00:00 UTC 2022
The SMYD3-MTHFD1L-formate metabolic regulatory axis mediates mitophagy to inhibit M1 polarization in macrophages.
International immunopharmacology
Zhu W,Wang C,Xue L,Liu L,Yang X,Liu Z,Zhang S,Luo D
Thu Dec 01 00:00:00 UTC 2022
The SMYD3-MTHFD1L-formate metabolic regulatory axis mediates mitophagy to inhibit M1 polarization in macrophages.
International immunopharmacology
Zhu W,Wang C,Xue L,Liu L,Yang X,Liu Z,Zhang S,Luo D
Thu Dec 01 00:00:00 UTC 2022
Autophagy-associated circRNA circATG7 facilitates autophagy and promotes pancreatic cancer progression.
Cell death & disease
He Z,Cai K,Zeng Z,Lei S,Cao W,Li X
Mon Mar 14 00:00:00 UTC 2022
Autophagy-associated circRNA circATG7 facilitates autophagy and promotes pancreatic cancer progression.
Cell death & disease
He Z,Cai K,Zeng Z,Lei S,Cao W,Li X
Mon Mar 14 00:00:00 UTC 2022
Autophagy-associated circRNA circATG7 facilitates autophagy and promotes pancreatic cancer progression.
Cell death & disease
He Z,Cai K,Zeng Z,Lei S,Cao W,Li X
Mon Mar 14 00:00:00 UTC 2022
Autophagy-associated circRNA circATG7 facilitates autophagy and promotes pancreatic cancer progression.
Cell death & disease
He Z,Cai K,Zeng Z,Lei S,Cao W,Li X
Mon Mar 14 00:00:00 UTC 2022
Autophagy-associated circRNA circATG7 facilitates autophagy and promotes pancreatic cancer progression.
Cell death & disease
He Z,Cai K,Zeng Z,Lei S,Cao W,Li X
Mon Mar 14 00:00:00 UTC 2022
Autophagy-associated circRNA circATG7 facilitates autophagy and promotes pancreatic cancer progression.
Cell death & disease
He Z,Cai K,Zeng Z,Lei S,Cao W,Li X
Mon Mar 14 00:00:00 UTC 2022
Transcriptome Analysis Reveal Candidate Genes and Pathways Responses to Lactate Dehydrogenase Inhibition (Oxamate) in Hyperglycemic Human Renal Proximal Epithelial Tubular Cells.
Frontiers in endocrinology
Wang Z,Hao D,Fang D,Yu J,Wang X,Qin G
Tue Apr 05 00:00:00 UTC 2022
GPR43 activation-mediated lipotoxicity contributes to podocyte injury in diabetic nephropathy by modulating the ERK/EGR1 pathway.
International journal of biological sciences
Lu J,Chen PP,Zhang JX,Li XQ,Wang GH,Yuan BY,Huang SJ,Liu XQ,Jiang TT,Wang MY,Liu WT,Ruan XZ,Liu BC,Ma KL
Mon Mar 28 00:00:00 UTC 2022
GPR43 activation-mediated lipotoxicity contributes to podocyte injury in diabetic nephropathy by modulating the ERK/EGR1 pathway.
International journal of biological sciences
Lu J,Chen PP,Zhang JX,Li XQ,Wang GH,Yuan BY,Huang SJ,Liu XQ,Jiang TT,Wang MY,Liu WT,Ruan XZ,Liu BC,Ma KL
Mon Mar 28 00:00:00 UTC 2022
Caryophyllene Oxide Induces Ferritinophagy by Regulating the NCOA4/FTH1/LC3 Pathway in Hepatocellular Carcinoma.
Frontiers in pharmacology
Xiu Z,Zhu Y,Han J,Li Y,Yang X,Yang G,Song G,Li S,Li Y,Cheng C,Li Y,Fang J,Li X,Jin N
Fri Jul 29 00:00:00 UTC 2022
Protection of catalpol against triptolide-induced hepatotoxicity by inhibiting excessive autophagy via the PERK-ATF4-CHOP pathway.
PeerJ
Zhang L,Li C,Fu L,Yu Z,Xu G,Zhou J,Shen M,Feng Z,Zhu H,Xie T,Zhou L,Zhou X
Fri Jan 06 00:00:00 UTC 2023
Calcitriol-enhanced autophagy in gingival epithelium attenuates periodontal inflammation in rats with type 2 diabetes mellitus.
Frontiers in endocrinology
Wang Y,Huang M,Xu W,Li F,Ma C,Tang X
Mon Jan 30 00:00:00 UTC 2023
Calcitriol-enhanced autophagy in gingival epithelium attenuates periodontal inflammation in rats with type 2 diabetes mellitus.
Frontiers in endocrinology
Wang Y,Huang M,Xu W,Li F,Ma C,Tang X
Mon Jan 30 00:00:00 UTC 2023
A neutrophil-mediated carrier regulates tumor stemness by inhibiting autophagy to prevent postoperative triple-negative breast cancer recurrence and metastasis.
Acta biomaterialia
Ren K,He J,Qiu Y,Xu Z,Wang X,Li J,Zang S,Yang Y,Li J,Long Y,Zhang Z,Li M,He Q
Wed Jun 01 00:00:00 UTC 2022
Remote cyclic compression ameliorates myocardial infarction injury in rats via AMPK-dependent pathway.
Microvascular research
Xu S,Xia X,Liu Y,Chen F,Gu R,Bian X,Xu X,Jia C,Lu S,Gu Y,Bai H,Zhang H
Sun May 01 00:00:00 UTC 2022
Long non-coding RNA IGFBP7-AS1 promotes odontogenic differentiation of stem cells from human exfoliated deciduous teeth through autophagy: An in vitro study.
Archives of oral biology
Wang D,Zhu N,Xie F,Qin M,Wang Y
Thu Sep 01 00:00:00 UTC 2022
Overexpression of VPS16 correlates with tumor progression and chemoresistance in colorectal cancer.
Biochemical and biophysical research communications
Zhang B,Ma Y,Niu H,Liu Z
Sat Jun 04 00:00:00 UTC 2022
Autophagy Inhibition Enhances the Anti-Tumor Activity of Methylseleninic Acid in Cisplatin-Resistance Human Lung Adenocarcinoma Cells.
Frontiers in pharmacology
Xin M,Gao Q,Xiang X,Xu J,Jiao Y,Li X,Zhang X,Jia X
Wed Nov 01 00:00:00 UTC 2023
Repeated social defeat stress inhibits development of hippocampus neurons through mitophagy and autophagy.
Brain research bulletin
Guo L,Jiang ZM,Sun RX,Pang W,Zhou X,Du ML,Chen MX,Lv X,Wang JT
Sun May 01 00:00:00 UTC 2022
Short-term starvation synergistically enhances cytotoxicity of Niraparib via Akt/mTOR signaling pathway in ovarian cancer therapy.
Cancer cell international
Zhi W,Li S,Wan Y,Wu F,Hong L
Tue Jan 11 00:00:00 UTC 2022
mTORC1 Mediates the Processes of Lysine Regulating Satellite Cells Proliferation, Apoptosis, and Autophagy.
Metabolites
Liu M,Yue Z,Zhang B,Li F,Liu L,Li F
Thu Aug 25 00:00:00 UTC 2022
Structural Variability of Lipoarabinomannan Modulates Innate Immune Responses within Infected Alveolar Epithelial Cells.
Cells
Liu H,Gui X,Chen S,Fu W,Li X,Xiao T,Hou J,Jiang T
Fri Jan 21 00:00:00 UTC 2022
Vitamin D3 promotes autophagy in THP-1 cells infected with Mycobacterium tuberculosis.
Experimental and therapeutic medicine
Wu Y,Lin X,Song F,Xue D,Wang Y
Tue Mar 01 00:00:00 UTC 2022
Effect of rapamycin treatment on oocyte in vitro maturation and embryonic development after parthenogenesis in yaks.
Theriogenology
Zhang T,Wang L,Pan Y,He H,Wang J,Zhao T,Ding T,Wang Y,Zhao L,Han X,Fan J,Xu G,Cui Y,Yu S
Tue Nov 01 00:00:00 UTC 2022
Harpagoside Protects Against Doxorubicin-Induced Cardiotoxicity via P53-Parkin-Mediated Mitophagy.
Frontiers in cell and developmental biology
Li W,Wang X,Liu T,Zhang Q,Cao J,Jiang Y,Sun Q,Li C,Wang W,Wang Y
Tue Mar 01 00:00:00 UTC 2022
Rosa rugosa polysaccharide induces autophagy-mediated apoptosis in human cervical cancer cells via the PI3K/AKT/mTOR pathway.
International journal of biological macromolecules
Liu Y,Li H,Zheng Z,Niu A,Liu S,Li W,Ren P,Liu Y,Inam M,Guan L,Ma H
Fri Jul 01 00:00:00 UTC 2022
Purple sweet potato delphinidin-3-rutin represses glioma proliferation by inducing miR-20b-5p/Atg7-dependent cytostatic autophagy.
Molecular therapy oncolytics
Wang M,Liu K,Bu H,Cong H,Dong G,Xu N,Li C,Zhao Y,Jiang F,Zhang Y,Yuan B,Li R,Jiang J
Thu Sep 15 00:00:00 UTC 2022
Effects of ferulic acid, a major component of rice bran, on proliferation, apoptosis, and autophagy of HepG2 cells.
Food research international (Ottawa, Ont.)
Wang J,Lai X,Yuan D,Liu Y,Wang J,Liang Y
Tue Nov 01 00:00:00 UTC 2022
Overexpression of sFlt-1 represses ox-LDL-induced injury of HUVECs by activating autophagy via PI3K/AKT/mTOR pathway.
Microvascular research
Zhou YH,Tang YZ,Guo LY,Zheng LL,Zhang D,Yang CY,Wang W
Sat Jan 01 00:00:00 UTC 2022
Deglycosylated EpCAM regulates proliferation by enhancing autophagy of breast cancer cells via PI3K/Akt/mTOR pathway.
Aging
Yang L,Wang Q,Zhao Q,Yang F,Liu T,Huang X,Yan Q,Yang X
Tue Jan 04 00:00:00 UTC 2022
GCAF(TMEM251) regulates lysosome biogenesis by activating the mannose-6-phosphate pathway.
Nature communications
Zhang W,Yang X,Li Y,Yu L,Zhang B,Zhang J,Cho WJ,Venkatarangan V,Chen L,Burugula BB,Bui S,Wang Y,Duan C,Kitzman JO,Li M
Mon Sep 12 00:00:00 UTC 2022
Thioredoxin upregulation delays diabetes-induced photoreceptor cell degeneration via AMPK-mediated autophagy and exosome secretion.
Diabetes research and clinical practice
Ren X,Lv J,Wang N,Liu J,Gao C,Wu X,Yu Y,Teng Q,Dong W,Kong H,Kong L
Tue Mar 01 00:00:00 UTC 2022
Thioredoxin upregulation delays diabetes-induced photoreceptor cell degeneration via AMPK-mediated autophagy and exosome secretion.
Diabetes research and clinical practice
Ren X,Lv J,Wang N,Liu J,Gao C,Wu X,Yu Y,Teng Q,Dong W,Kong H,Kong L
Tue Mar 01 00:00:00 UTC 2022
Parkin Inhibits RANKL-Induced Osteoclastogenesis and Ovariectomy-Induced Bone Loss.
Biomolecules
Hou W,Chen M,Ye C,Chen E,Li W,Zhang W
Mon Oct 31 00:00:00 UTC 2022
Design, synthesis, and biological evaluation of novel dual inhibitors of heat shock protein 90/mammalian target of rapamycin (Hsp90/mTOR) against bladder cancer cells.
European journal of medicinal chemistry
Pan Z,Chen Y,Pang H,Wang X,Zhang Y,Xie X,He G
Tue Nov 15 00:00:00 UTC 2022
Design, synthesis, and biological evaluation of novel dual inhibitors of heat shock protein 90/mammalian target of rapamycin (Hsp90/mTOR) against bladder cancer cells.
European journal of medicinal chemistry
Pan Z,Chen Y,Pang H,Wang X,Zhang Y,Xie X,He G
Tue Nov 15 00:00:00 UTC 2022
Design, synthesis, and biological evaluation of novel dual inhibitors of heat shock protein 90/mammalian target of rapamycin (Hsp90/mTOR) against bladder cancer cells.
European journal of medicinal chemistry
Pan Z,Chen Y,Pang H,Wang X,Zhang Y,Xie X,He G
Tue Nov 15 00:00:00 UTC 2022
Neutrophil extracellular traps promote thrombogenicity in cerebral venous sinus thrombosis.
Cell & bioscience
Jin J,Qiao S,Liu J,Li W,Wang F,Gao X,Tian J,Wang N,Zhang J,Dong J,Li H,Wang J,Hu S,Zhou P
Fri Jul 22 00:00:00 UTC 2022
Xuesaitong Combined with Dexmedetomidine Improves Cerebral Ischemia-Reperfusion Injury in Rats by Activating Keap1/Nrf2 Signaling and Mitophagy in Hippocampal Tissue.
Oxidative medicine and cellular longevity
Han GJ,Min XZ,Ma SS,Ding C,Wang XQ
Tue Dec 20 00:00:00 UTC 2022
PERK Regulates the Sensitivity of Hepatocellular Carcinoma Cells to High-LET Carbon Ions via either Apoptosis or Ferroptosis.
Journal of Cancer
Zheng X,Liu B,Liu X,Li P,Zhang P,Ye F,Zhao T,Kuang Y,Chen W,Jin X,Li Q
Tue Jan 25 00:00:00 UTC 2022
LncRNA MEG3 promotes the sensitivity of bortezomib by inhibiting autophagy in multiple myeloma.
Leukemia research
Zang X,Wang J,Xia Y,Li J,Chen L,Gu Y,Shen X
Thu Dec 01 00:00:00 UTC 2022
Inhibition of HSPA8 by rifampicin contributes to ferroptosis via enhancing autophagy.
Liver international : official journal of the International Association for the Study of the Liver
Zhou J,Tan Y,Hu L,Fu J,Li D,Chen J,Long Y
Thu Dec 01 00:00:00 UTC 2022
Inhibition of HSPA8 by rifampicin contributes to ferroptosis via enhancing autophagy.
Liver international : official journal of the International Association for the Study of the Liver
Zhou J,Tan Y,Hu L,Fu J,Li D,Chen J,Long Y
Thu Dec 01 00:00:00 UTC 2022
Inhibition of HSPA8 by rifampicin contributes to ferroptosis via enhancing autophagy.
Liver international : official journal of the International Association for the Study of the Liver
Zhou J,Tan Y,Hu L,Fu J,Li D,Chen J,Long Y
Thu Dec 01 00:00:00 UTC 2022
Inhibition of HSPA8 by rifampicin contributes to ferroptosis via enhancing autophagy.
Liver international : official journal of the International Association for the Study of the Liver
Zhou J,Tan Y,Hu L,Fu J,Li D,Chen J,Long Y
Thu Dec 01 00:00:00 UTC 2022
LncRNA CCAT1 Upregulates ATG5 to Enhance Autophagy and Promote Gastric Cancer Development by Absorbing miR-140-3p.
Digestive diseases and sciences
Yang F,Peng ZX,Ji WD,Yu JD,Qian C,Liu JD,Fang GE
Mon Aug 01 00:00:00 UTC 2022
Deciphering the Mechanism of Bushen Huoxue Decotion on Decidualization by Intervening Autophagy via AMPK/mTOR/ULK1: A Novel Discovery for URSA Treatment.
Frontiers in pharmacology
Zhao X,Jiang Y,Ren J,Wang Y,Zhao Y,Feng X
Wed Feb 16 00:00:00 UTC 2022
Deciphering the Mechanism of Bushen Huoxue Decotion on Decidualization by Intervening Autophagy via AMPK/mTOR/ULK1: A Novel Discovery for URSA Treatment.
Frontiers in pharmacology
Zhao X,Jiang Y,Ren J,Wang Y,Zhao Y,Feng X
Wed Feb 16 00:00:00 UTC 2022
Calycosin induces autophagy and apoptosis via Sestrin2/AMPK/mTOR in human papillary thyroid cancer cells.
Frontiers in pharmacology
Qu N,Qu J,Huang N,Zhang K,Ye T,Shi J,Chen B,Kan C,Zhang J,Han F,Hou N,Sun X,Pan R
Tue Jan 03 00:00:00 UTC 2023
SIRT1/mTOR pathway-mediated autophagy dysregulation promotes Pb-induced hepatic lipid accumulation in HepG2 cells.
Environmental toxicology
Zhao Y,Mao A,Zhang R,Guan S,Lu J
Tue Mar 01 00:00:00 UTC 2022
Achyranthoside D (AD) improve intervertebral disc degeneration through affect the autophagy and the activation of PI3K/Akt/mTOR pathway.
Journal of orthopaedic surgery (Hong Kong)
Zhang C,Li Z,Li L,Li S,Yang L,Chen L,Zhang X,Yang S,Guo Y
Tue Nov 01 00:00:00 UTC 2022
Achyranthoside D (AD) improve intervertebral disc degeneration through affect the autophagy and the activation of PI3K/Akt/mTOR pathway.
Journal of orthopaedic surgery (Hong Kong)
Zhang C,Li Z,Li L,Li S,Yang L,Chen L,Zhang X,Yang S,Guo Y
Tue Nov 01 00:00:00 UTC 2022
MiR-152-5p suppresses osteogenic differentiation of mandible mesenchymal stem cells by regulating ATG14-mediated autophagy.
Stem cell research & therapy
Li S,Gao L,Zhang W,Yu Y,Zheng J,Liang X,Xin S,Ren W,Zhi K
Tue Jul 26 00:00:00 UTC 2022
MiR-152-5p suppresses osteogenic differentiation of mandible mesenchymal stem cells by regulating ATG14-mediated autophagy.
Stem cell research & therapy
Li S,Gao L,Zhang W,Yu Y,Zheng J,Liang X,Xin S,Ren W,Zhi K
Tue Jul 26 00:00:00 UTC 2022
Ursolic Acid Derivative UA232 Promotes Tumor Cell Apoptosis by Inducing Endoplasmic Reticulum Stress and Lysosomal Dysfunction.
International journal of biological sciences
Gou W,Luo N,Yu B,Wu H,Wu S,Tian C,Guo J,Ning H,Bi C,Wei H,Hou W,Li Y
Thu Apr 14 00:00:00 UTC 2022
Ursolic Acid Derivative UA232 Promotes Tumor Cell Apoptosis by Inducing Endoplasmic Reticulum Stress and Lysosomal Dysfunction.
International journal of biological sciences
Gou W,Luo N,Yu B,Wu H,Wu S,Tian C,Guo J,Ning H,Bi C,Wei H,Hou W,Li Y
Thu Apr 14 00:00:00 UTC 2022
3,4,5-O-tricaffeoylquinic acid with anti-radiation activity suppresses LPS-induced NLRP3 inflammasome activation via autophagy in THP-1 macrophages.
Molecular immunology
Liu J,Chen J,Xu B,Lin L,Liu S,Ma X,Liu J
Fri Jul 01 00:00:00 UTC 2022
Magnetic hyperthermia induces effective and genuine immunogenic tumor cell death with respect to exogenous heating.
Journal of materials chemistry. B
Yan B,Liu C,Wang S,Li H,Jiao J,Lee WSV,Zhang S,Hou Y,Hou Y,Ma X,Fan H,Lv Y,Liu X
Wed Jul 20 00:00:00 UTC 2022
Poloxamer 407 and Hyaluronic Acid Thermosensitive Hydrogel-Encapsulated Ginsenoside Rg3 to Promote Skin Wound Healing.
Frontiers in bioengineering and biotechnology
Peng X,Ding C,Zhao Y,Hao M,Liu W,Yang M,Xiao F,Zheng Y
Sat Jul 23 00:00:00 UTC 2022
Mitochondrial dysfunction is involved in the cellular activity inhibition by eleutheroside B in SMMC-7721 and HeLa cells.
Human & experimental toxicology
Sun S,Zhang Y,Xu W,Wang B,Chen J
Thu May 19 00:00:00 UTC 2022
Grass Carp Reovirus triggers autophagy enhancing virus replication via the Akt/mTOR pathway.
Fish & shellfish immunology
Zhu M,Zhang Y,Pan J,Tong X,Zhang X,Hu X,Gong C
Thu Sep 01 00:00:00 UTC 2022
Mitochondrial membrane proteins and VPS35 orchestrate selective removal of mtDNA.
Nature communications
Sen A,Kallabis S,Gaedke F,Jüngst C,Boix J,Nüchel J,Maliphol K,Hofmann J,Schauss AC,Krüger M,Wiesner RJ,Pla-Martín D
Mon Nov 07 00:00:00 UTC 2022
Mitochondrial membrane proteins and VPS35 orchestrate selective removal of mtDNA.
Nature communications
Sen A,Kallabis S,Gaedke F,Jüngst C,Boix J,Nüchel J,Maliphol K,Hofmann J,Schauss AC,Krüger M,Wiesner RJ,Pla-Martín D
Mon Nov 07 00:00:00 UTC 2022
Asymptomatic Hyperuricemia Is Associated with Achilles Tendon Rupture through Disrupting the Normal Functions of Tendon Stem/Progenitor Cells.
Stem cells international
Liang J,Chen B,Li Y,Nie D,Lei C,Yang Q,Zhou Y,Li S,Chen S,Li B,Shu L,Chen L,Wang W
Thu Dec 22 00:00:00 UTC 2022
Inhibition of autophagy potentiates the cytotoxicity of the irreversible FGFR1-4 inhibitor FIIN-2 on lung adenocarcinoma.
Cell death & disease
Jia X,Xin M,Xu J,Xiang X,Li X,Jiao Y,Wang L,Jiang J,Pang F,Zhang X,Zhang J
Tue Aug 30 00:00:00 UTC 2022
FUNDC1-mediated mitophagy and HIF1α activation drives pulmonary hypertension during hypoxia.
Cell death & disease
Liu R,Xu C,Zhang W,Cao Y,Ye J,Li B,Jia S,Weng L,Liu Y,Liu L,Zheng M
Thu Jul 21 00:00:00 UTC 2022
BRD4 Inhibition Suppresses Senescence and Apoptosis of Nucleus Pulposus Cells by Inducing Autophagy during Intervertebral Disc Degeneration: An In Vitro and In Vivo Study.
Oxidative medicine and cellular longevity
Zhang GZ,Chen HW,Deng YJ,Liu MQ,Wu ZL,Ma ZJ,He XG,Gao YC,Kang XW
Mon Apr 04 00:00:00 UTC 2022
Comprehensive analysis of placental gene-expression profiles and identification of EGFR-mediated autophagy and ferroptosis suppression in intrahepatic cholestasis of pregnancy.
Gene
Fang Y,Fang D
Fri Aug 05 00:00:00 UTC 2022
Epigallocatechin-3-gallate attenuates myocardial fibrosis in diabetic rats by activating autophagy.
Experimental biology and medicine (Maywood, N.J.)
Jia Q,Yang R,Mehmood S,Li Y
Thu Sep 01 00:00:00 UTC 2022
miR-146a-5p/TXNIP axis attenuates intestinal ischemia-reperfusion injury by inhibiting autophagy via the PRKAA/mTOR signaling pathway.
Biochemical pharmacology
Zhenzhen L,Wenting L,Jianmin Z,Guangru Z,Disheng L,Zhiyu Z,Feng C,Yajing S,Yingxiang H,Jipeng L,Zhanhai W,Yan Z,Xin L,Yongqiang L,Yufang L
Tue Mar 01 00:00:00 UTC 2022
Long-Term Consumption of Food-Derived Chlorogenic Acid Protects Mice against Acetaminophen-Induced Hepatotoxicity via Promoting PINK1-Dependent Mitophagy and Inhibiting Apoptosis.
Toxics
Hu B,Li J,Gong D,Dai Y,Wang P,Wan L,Xu S
Sat Nov 05 00:00:00 UTC 2022
Long-Term Consumption of Food-Derived Chlorogenic Acid Protects Mice against Acetaminophen-Induced Hepatotoxicity via Promoting PINK1-Dependent Mitophagy and Inhibiting Apoptosis.
Toxics
Hu B,Li J,Gong D,Dai Y,Wang P,Wan L,Xu S
Sat Nov 05 00:00:00 UTC 2022
Polyvinylpyrrolidone-Modified Taxifolin Liposomes Promote Liver Repair by Modulating Autophagy to Inhibit Activation of the TLR4/NF-κB Signaling Pathway.
Frontiers in bioengineering and biotechnology
Ding Q,Liu W,Liu X,Ding C,Zhao Y,Dong L,Chen H,Sun S,Zhang Y,Zhang J,Wu M
Sat Jul 16 00:00:00 UTC 2022
C9orf72 knockdown alleviates hepatic insulin resistance by promoting lipophagy.
Biochemical and biophysical research communications
Cang X,Wang Y,Zeng J,Gao J,Yu Q,Lu C,Xu F,Lin J,Zhu J,Wang X
Sat Jan 15 00:00:00 UTC 2022
C9orf72 knockdown alleviates hepatic insulin resistance by promoting lipophagy.
Biochemical and biophysical research communications
Cang X,Wang Y,Zeng J,Gao J,Yu Q,Lu C,Xu F,Lin J,Zhu J,Wang X
Sat Jan 15 00:00:00 UTC 2022
Resveratrol alleviates enterotoxigenic Escherichia coli K88-induced damage by regulating SIRT-1 signaling in intestinal porcine epithelial cells.
Food & function
Luo X,Wu S,Jia H,Si X,Song Z,Zhai Z,Bai J,Li J,Yang Y,Wu Z
Mon Jul 04 00:00:00 UTC 2022
Activating PPARβ/δ Protects against Endoplasmic Reticulum Stress-Induced Astrocytic Apoptosis via UCP2-Dependent Mitophagy in Depressive Model.
International journal of molecular sciences
Ji J,Li S,Jiang Z,Yu J,Sun Y,Cai Z,Dong Y,Sun X
Fri Sep 16 00:00:00 UTC 2022
TMT-Based Quantitative Proteomics Analysis Reveals the Panoramic Pharmacological Molecular Mechanism of β-Elemonic Acid Inhibition of Colorectal Cancer.
Frontiers in pharmacology
Xia Y,Yang J,Li C,Hao X,Fan H,Zhao Y,Tang J,Wan X,Lian S,Yang J
Sat Mar 05 00:00:00 UTC 2022
Rab21 Protein Is Degraded by Both the Ubiquitin-Proteasome Pathway and the Autophagy-Lysosome Pathway.
International journal of molecular sciences
Liu P,Wu A,Li H,Zhang J,Ni J,Quan Z,Qing H
Thu Jan 20 00:00:00 UTC 2022
Rab21 Protein Is Degraded by Both the Ubiquitin-Proteasome Pathway and the Autophagy-Lysosome Pathway.
International journal of molecular sciences
Liu P,Wu A,Li H,Zhang J,Ni J,Quan Z,Qing H
Thu Jan 20 00:00:00 UTC 2022
Haizao Yuhu decoctions including three species of glycyrrhiza protected against propylthiouracil-induced goiter with hypothyroidism in rats via the AMPK/mTOR pathway.
Journal of ethnopharmacology
Li N,Li M,Xiu L,Liao W,Ren Y,Liu H,Chen S,Chen F,Yu X,Fan A,Huo M,He J,Zhong G
Wed Oct 05 00:00:00 UTC 2022
Haizao Yuhu decoctions including three species of glycyrrhiza protected against propylthiouracil-induced goiter with hypothyroidism in rats via the AMPK/mTOR pathway.
Journal of ethnopharmacology
Li N,Li M,Xiu L,Liao W,Ren Y,Liu H,Chen S,Chen F,Yu X,Fan A,Huo M,He J,Zhong G
Wed Oct 05 00:00:00 UTC 2022
Colistin-induced pulmonary toxicity involves the activation of NOX4/TGF-β/mtROS pathway and the inhibition of Akt/mTOR pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Dai C,Li M,Sun T,Zhang Y,Wang Y,Shen Z,Velkov T,Tang S,Shen J
Sun May 01 00:00:00 UTC 2022
Src Activation Aggravates Podocyte Injury in Diabetic Nephropathy via Suppression of FUNDC1-Mediated Mitophagy.
Frontiers in pharmacology
Zheng T,Wang HY,Chen Y,Chen X,Wu ZL,Hu QY,Sun H
Sat Jul 16 00:00:00 UTC 2022
SUMO2-mediated SUMOylation of SH3GLB1 promotes ionizing radiation-induced hypertrophic cardiomyopathy through mitophagy activation.
European journal of pharmacology
Gao A,Zou J,Mao Z,Zhou H,Zeng G
Sun Jun 05 00:00:00 UTC 2022
Visualizing the native cellular organization by coupling cryofixation with expansion microscopy (Cryo-ExM).
Nature methods
Laporte MH,Klena N,Hamel V,Guichard P
Tue Feb 01 00:00:00 UTC 2022
Transcription factor nuclear factor erythroid 2 p45-related factor 2 (NRF2) ameliorates sepsis-associated acute kidney injury by maintaining mitochondrial homeostasis and improving the mitochondrial function.
European journal of histochemistry : EJH
Chen Z,Wang H,Hu B,Chen X,Zheng M,Liang L,Lyu J,Zeng Q
Tue Jun 21 00:00:00 UTC 2022
Transcription factor nuclear factor erythroid 2 p45-related factor 2 (NRF2) ameliorates sepsis-associated acute kidney injury by maintaining mitochondrial homeostasis and improving the mitochondrial function.
European journal of histochemistry : EJH
Chen Z,Wang H,Hu B,Chen X,Zheng M,Liang L,Lyu J,Zeng Q
Tue Jun 21 00:00:00 UTC 2022
Nobiletin mitigates NAFLD via lipophagy and inflammation.
Food & function
Yang X,Deng Y,Tu Y,Feng D,Liao W
Mon Oct 03 00:00:00 UTC 2022
Nobiletin mitigates NAFLD via lipophagy and inflammation.
Food & function
Yang X,Deng Y,Tu Y,Feng D,Liao W
Mon Oct 03 00:00:00 UTC 2022
Nobiletin mitigates NAFLD via lipophagy and inflammation.
Food & function
Yang X,Deng Y,Tu Y,Feng D,Liao W
Mon Oct 03 00:00:00 UTC 2022
Nobiletin mitigates NAFLD via lipophagy and inflammation.
Food & function
Yang X,Deng Y,Tu Y,Feng D,Liao W
Mon Oct 03 00:00:00 UTC 2022
An Experimental Study Reveals the Protective Effect of Autophagy against Realgar-Induced Liver Injury via Suppressing ROS-Mediated NLRP3 Inflammasome Pathway.
International journal of molecular sciences
Yang J,Li J,Guo H,Zhang Y,Guo Z,Liu Y,Huo T
Thu May 19 00:00:00 UTC 2022
GNIP1 functions both as a scaffold protein and an E3 ubiquitin ligase to regulate autophagy in lung cancer.
Cell communication and signaling : CCS
Zhou F,Liu Y,Ai W,Wang Y,Gan M,Jiang Q,Han T,Wang JB
Tue Aug 30 00:00:00 UTC 2022
GNIP1 functions both as a scaffold protein and an E3 ubiquitin ligase to regulate autophagy in lung cancer.
Cell communication and signaling : CCS
Zhou F,Liu Y,Ai W,Wang Y,Gan M,Jiang Q,Han T,Wang JB
Tue Aug 30 00:00:00 UTC 2022
Inhibiting autophagy flux and DNA repair of tumor cells to boost radiotherapy of orthotopic glioblastoma.
Biomaterials
Xu Q,Zhang H,Liu H,Han Y,Qiu W,Li Z
Sat Jan 01 00:00:00 UTC 2022
Catalpol Enhances Random-Pattern Skin Flap Survival by Activating SIRT1-Mediated Enhancement of Autophagy.
Oxidative medicine and cellular longevity
Jiang RH,Dong CJ,Chen ZL,Cheng S,Yang JX,Gao WY
Mon May 30 00:00:00 UTC 2022
Catalpol Enhances Random-Pattern Skin Flap Survival by Activating SIRT1-Mediated Enhancement of Autophagy.
Oxidative medicine and cellular longevity
Jiang RH,Dong CJ,Chen ZL,Cheng S,Yang JX,Gao WY
Mon May 30 00:00:00 UTC 2022
The microRNA-10b-Bim axis promotes cancer progression through activating autophagy in oral squamous cell carcinoma.
Cell death discovery
Li S,Gao L,Liu J,Guo C,Zheng J,Zhi K,Ren W
Thu Aug 25 00:00:00 UTC 2022
The microRNA-10b-Bim axis promotes cancer progression through activating autophagy in oral squamous cell carcinoma.
Cell death discovery
Li S,Gao L,Liu J,Guo C,Zheng J,Zhi K,Ren W
Thu Aug 25 00:00:00 UTC 2022
The microRNA-10b-Bim axis promotes cancer progression through activating autophagy in oral squamous cell carcinoma.
Cell death discovery
Li S,Gao L,Liu J,Guo C,Zheng J,Zhi K,Ren W
Thu Aug 25 00:00:00 UTC 2022
Magnesium isoglycyrrhizinate ameliorates lipopolysaccharide-induced liver injury by upregulating autophagy and inhibiting inflammation via IL-22 expression.
Bioorganic chemistry
Zhou X,Li X,Yi K,Liang C,Geng S,Zhu J,Xie C,Zhong C
Tue Nov 01 00:00:00 UTC 2022
Magnesium isoglycyrrhizinate ameliorates lipopolysaccharide-induced liver injury by upregulating autophagy and inhibiting inflammation via IL-22 expression.
Bioorganic chemistry
Zhou X,Li X,Yi K,Liang C,Geng S,Zhu J,Xie C,Zhong C
Tue Nov 01 00:00:00 UTC 2022
Magnesium isoglycyrrhizinate ameliorates lipopolysaccharide-induced liver injury by upregulating autophagy and inhibiting inflammation via IL-22 expression.
Bioorganic chemistry
Zhou X,Li X,Yi K,Liang C,Geng S,Zhu J,Xie C,Zhong C
Tue Nov 01 00:00:00 UTC 2022
Ethanol potentiates mirtazapine-induced cardiotoxicity by inducing dysfunctional autophagy via HMGB1-dependent Akt/mTOR signaling pathway.
Toxicology letters
Zhu R,Liu X,Xue Q,Dong X,Zhang T,Jiang Y
Fri Apr 01 00:00:00 UTC 2022
O-GlcNAcylation of SIRT1 Protects against Cold Stress-Induced Skeletal Muscle Damage via Amelioration of Mitochondrial Homeostasis.
International journal of molecular sciences
Cao Y,Zhang M,Li Y,Lu J,Zhou W,Li X,Shi H,Xu B,Li S
Tue Nov 22 00:00:00 UTC 2022
Maltol Promotes Mitophagy and Inhibits Oxidative Stress via the Nrf2/PINK1/Parkin Pathway after Spinal Cord Injury.
Oxidative medicine and cellular longevity
Mao Y,Du J,Chen X,Al Mamun A,Cao L,Yang Y,Mubwandarikwa J,Zaeem M,Zhang W,Chen Y,Dai Y,Xiao J,Ye K
Fri Mar 18 00:00:00 UTC 2022
Long noncoding RNA ZFAS1 suppresses osteogenic differentiation of bone marrow-derived mesenchymal stem cells by upregulating miR-499-EPHA5 axis.
Molecular and cellular endocrinology
Wu J,Lin T,Gao Y,Li X,Yang C,Zhang K,Wang C,Zhou X
Sat Jan 01 00:00:00 UTC 2022
Dietary folic acid supplementation improves semen quality and spermatogenesis through altering autophagy and histone methylation in the testis of aged broiler breeder roosters.
Theriogenology
Ye N,Lv Z,Huang Z,Cheng Y,Wei Q,Shi F
Tue Mar 15 00:00:00 UTC 2022
Oncostatin M receptor regulates osteoblast differentiation via extracellular signal-regulated kinase/autophagy signaling.
Stem cell research & therapy
Zhou J,Yang J,Dong Y,Shi Y,Zhu E,Yuan H,Li X,Wang B
Tue Jun 28 00:00:00 UTC 2022
MiR-371a-5p Positively Associates with Hepatocellular Carcinoma Malignancy but Sensitizes Cancer Cells to Oxaliplatin by Suppressing BECN1-Dependent Autophagy.
Life (Basel, Switzerland)
Lv Z,Qiu X,Jin P,Li Z,Zhang Y,Lv L,Song F
Thu Oct 20 00:00:00 UTC 2022
Protocatechualdehyde Rescues Oxygen-Glucose Deprivation/Reoxygenation-Induced Endothelial Cells Injury by Inducing Autophagy and Inhibiting Apoptosis via Regulation of SIRT1.
Frontiers in pharmacology
Cao S,Chen S,Qiao X,Guo Y,Liu F,Ding Z,Jin B
Sat May 07 00:00:00 UTC 2022
Bisphenol A impairs macrophages through inhibiting autophagy via AMPK/mTOR signaling pathway and inducing apoptosis.
Ecotoxicology and environmental safety
Wu M,Cong Y,Wang K,Yu H,Zhang X,Ma M,Duan Z,Pei X
Fri Apr 01 00:00:00 UTC 2022
Bisphenol A impairs macrophages through inhibiting autophagy via AMPK/mTOR signaling pathway and inducing apoptosis.
Ecotoxicology and environmental safety
Wu M,Cong Y,Wang K,Yu H,Zhang X,Ma M,Duan Z,Pei X
Fri Apr 01 00:00:00 UTC 2022
Rab11a promotes the malignant progression of ovarian cancer by inducing autophagy.
Genes & genomics
Wang Y,Ren Y,Li N,Zhao J,Zhao S
Tue Nov 01 00:00:00 UTC 2022
Rab11a promotes the malignant progression of ovarian cancer by inducing autophagy.
Genes & genomics
Wang Y,Ren Y,Li N,Zhao J,Zhao S
Tue Nov 01 00:00:00 UTC 2022
Tryptophan-sorbitol based carbon quantum dots for theranostics against hepatocellular carcinoma.
Journal of nanobiotechnology
Wang Y,Chen J,Tian J,Wang G,Luo W,Huang Z,Huang Y,Li N,Guo M,Fan X
Mon Feb 14 00:00:00 UTC 2022
P-Glycoprotein Aggravates Blood Brain Barrier Dysfunction in Experimental Ischemic Stroke by Inhibiting Endothelial Autophagy.
Aging and disease
Huang L,Chen Y,Liu R,Li B,Fei X,Li X,Liu G,Li Y,Xu B,Fang W
Sat Oct 01 00:00:00 UTC 2022
An oleanolic acid derivative, K73-03, inhibits pancreatic cancer cells proliferation in vitro and in vivo via blocking EGFR/Akt pathway.
Cell biology international
Zhou Z,Dong Y,Li N,Niu M,Wang S,Zhou Y,Sun Z,Chu P,Tang Z
Tue Nov 01 00:00:00 UTC 2022
Exercise activates autophagy and regulates endoplasmic reticulum stress in muscle of high-fat diet mice to alleviate insulin resistance.
Biochemical and biophysical research communications
Cheng F,Dun Y,Cheng J,Ripley-Gonzalez JW,Jiang W,You B,Liu S
Sat Apr 23 00:00:00 UTC 2022
Integrated metabolism and epigenetic modifications in the macrophages of mice in responses to cold stress.
Journal of Zhejiang University. Science. B
Lu J,Fu S,Dai J,Hu J,Li S,Ji H,Wang Z,Yu J,Bao J,Xu B,Guo J,Yang H
Wed Jun 15 00:00:00 UTC 2022
Paired related homeobox 1 attenuates autophagy via acetyl-CoA carboxylase 1-regulated fatty acid metabolism in salivary adenoid cystic carcinoma.
FEBS open bio
Gao J,Ma K,Zhang L,Li T,Zhao B,Jiang Y
Sun May 01 00:00:00 UTC 2022
Multifunctional theranostic nanoplatform loaded with autophagy inhibitor for enhanced photothermal cancer therapy under mild near-infrared irradiation.
Biomaterials advances
Shi M,Liu Y,Huang J,Chen Z,Ni C,Lu J,Zhang Y,Liu Z,Bai J
Fri Jul 01 00:00:00 UTC 2022
Dye-Sensitized Rare Earth Nanoparticles with Up/Down Conversion Luminescence for On-Demand Gas Therapy of Glioblastoma Guided by NIR-II Fluorescence Imaging.
Advanced healthcare materials
Liu Z,Yun B,Han Y,Jiang Z,Zhu H,Ren F,Li Z
Tue Feb 01 00:00:00 UTC 2022
Combination of polythyleneimine regulating autophagy prodrug and Mdr1 siRNA for tumor multidrug resistance.
Journal of nanobiotechnology
Wang C,Li Z,Xu P,Xu L,Han S,Sun Y
Fri Nov 11 00:00:00 UTC 2022
Combination of polythyleneimine regulating autophagy prodrug and Mdr1 siRNA for tumor multidrug resistance.
Journal of nanobiotechnology
Wang C,Li Z,Xu P,Xu L,Han S,Sun Y
Fri Nov 11 00:00:00 UTC 2022
Aberrant SPOP-CHAF1A ubiquitination axis triggers tumor autophagy that endows a therapeutical vulnerability in diffuse large B cell lymphoma.
Journal of translational medicine
Yan W,Shi X,Wang H,Liao A,Yang W
Thu Jun 30 00:00:00 UTC 2022
Hsa_circRNA_0008028 Deficiency Ameliorates High Glucose-Induced Proliferation, Calcification, and Autophagy of Vascular Smooth Muscle Cells via miR-182-5p/TRIB3 Axis.
Oxidative medicine and cellular longevity
Shi L,Li Y,Shi M,Li X,Li G,Cen J,Liu D,Wei C,Lin Y
Wed Sep 07 00:00:00 UTC 2022
MiR-7-5p suppresses invasion via downregulation of the autophagy-related gene ATG7 and increases chemoresistance to cisplatin in BCa.
Bioengineered
Wang C,Tang Z,Zhang Z,Liu T,Zhang J,Huang H,Li Y
Tue Mar 01 00:00:00 UTC 2022
Estrogen-related Receptor α (ERRα) Functions in The Hypoxic Injury of Microglial Cells.
Journal of veterinary research
Deng CY,Zhu TT,Lian S,Wang JF,Wu R,Zheng JS
Tue Mar 01 00:00:00 UTC 2022
Evaluation of BPA effects on autophagy in Neuro-2a cells.
Toxicology and industrial health
Li J,Yin Z,Hua L,Wang X,Ren F,Ge Y
Tue Mar 01 00:00:00 UTC 2022
The cardiotoxicity of asthmatic rats after traffic-related PM2.5 and water-soluble components exposure mediated by endoplasmic reticulum stress and autophagy.
Environmental science and pollution research international
Tian J,Shi H,Wang X,Wu H,Wang C,Liu N,Wang D,Shi D,Zhang H,Zhang Z
Fri Jul 01 00:00:00 UTC 2022
Astragaloside IV Protects Detrusor from Partial Bladder Outlet Obstruction-Induced Oxidative Stress by Activating Mitophagy through AMPK-ULK1 Pathway.
Oxidative medicine and cellular longevity
Zhu X,Tao F,Zhang L,Yu Y,Xu Y,Yang G,Wei Z,Cheng Y,Yin X,Zhang X,Wei W,Wang A
Tue Jul 26 00:00:00 UTC 2022
Sodium butyrate attenuates bovine mammary epithelial cell injury by inhibiting the formation of neutrophil extracellular traps.
International immunopharmacology
Li Y,Liu J,Cui Y,Cao Y,Xu P,Kan X,Guo W,Fu S
Thu Sep 01 00:00:00 UTC 2022
The selective NLRP3 inflammasome inhibitor MCC950 improves isoproterenol-induced cardiac dysfunction by inhibiting cardiomyocyte senescence.
European journal of pharmacology
Shi Y,Zhao L,Wang J,Liu S,Zhang Y,Qin Q
Thu Dec 15 00:00:00 UTC 2022
Comprehensive Analysis of Quantitative Proteomics With DIA Mass Spectrometry and ceRNA Network in Intrahepatic Cholestasis of Pregnancy.
Frontiers in cell and developmental biology
Fang D,Fang Y,Zhang W,Xiang Y,Cheng X,Liang M,Xia H
Tue Aug 09 00:00:00 UTC 2022
Cinobufagin induces acute promyelocytic leukaemia cell apoptosis and PML-RARA degradation in a caspase-dependent manner by inhibiting the β-catenin signalling pathway.
Pharmaceutical biology
Bian Y,Xue M,Guo X,Jiang W,Zhao Y,Zhang Z,Wang X,Hu Y,Zhang Q,Dun W,Zhang L
Thu Dec 01 00:00:00 UTC 2022
A missing piece of the puzzle in pulmonary fibrosis: anoikis resistance promotes fibroblast activation.
Cell & bioscience
Yin J,Wang J,Zhang X,Liao Y,Luo W,Wang S,Ding J,Huang J,Chen M,Wang W,Fang S,Chao J
Fri Feb 25 00:00:00 UTC 2022
Systematic evaluation of the anti-tumor effect of Phellinus linteus polysaccharide in thyroid carcinoma in vitro.
Molecular biology reports
Yu K,Tan Z,Xin Y
Fri Apr 01 00:00:00 UTC 2022
Autophagic degradation of claudin-5 mediated by its binding to a Clostridium perfringens enterotoxin fragment modulates endothelial barrier permeability.
FEBS letters
Lin P,Tan R,Yu P,Li Y,Mo Y,Li W,Zhang J
Fri Apr 01 00:00:00 UTC 2022
The oncoprotein MUC1 facilitates breast cancer progression by promoting Pink1-dependent mitophagy via ATAD3A destabilization.
Cell death & disease
Li Q,Chu Y,Li S,Yu L,Deng H,Liao C,Liao X,Yang C,Qi M,Cheng J,Chen G,Huang L
Wed Oct 26 00:00:00 UTC 2022
Melatonin alleviates renal injury by activating mitophagy in diabetic nephropathy.
Frontiers in endocrinology
Tang H,Yang M,Liu Y,Zhu X,Liu S,Liu H,Sun L,Song P
Tue Aug 23 00:00:00 UTC 2022
Melatonin alleviates renal injury by activating mitophagy in diabetic nephropathy.
Frontiers in endocrinology
Tang H,Yang M,Liu Y,Zhu X,Liu S,Liu H,Sun L,Song P
Tue Aug 23 00:00:00 UTC 2022
Targeting CRL4 suppresses chemoresistant ovarian cancer growth by inducing mitophagy.
Signal transduction and targeted therapy
Meng Y,Qiu L,Zeng X,Hu X,Zhang Y,Wan X,Mao X,Wu J,Xu Y,Xiong Q,Chen Z,Zhang B,Han J
Fri Dec 09 00:00:00 UTC 2022
Targeting CRL4 suppresses chemoresistant ovarian cancer growth by inducing mitophagy.
Signal transduction and targeted therapy
Meng Y,Qiu L,Zeng X,Hu X,Zhang Y,Wan X,Mao X,Wu J,Xu Y,Xiong Q,Chen Z,Zhang B,Han J
Fri Dec 09 00:00:00 UTC 2022
The role of APTX4870 peptide in reducing cellular inflammatory responses by inhibiting Mycobacterium tuberculosis-derived mycolic acid-induced cytotoxicity.
Frontiers in microbiology
Lin X,Jia W,Feng G,Su Y,Kang Y,Zhang C,Liu W,Lu Z,Xue D
Fri Nov 11 00:00:00 UTC 2022
The role of APTX4870 peptide in reducing cellular inflammatory responses by inhibiting Mycobacterium tuberculosis-derived mycolic acid-induced cytotoxicity.
Frontiers in microbiology
Lin X,Jia W,Feng G,Su Y,Kang Y,Zhang C,Liu W,Lu Z,Xue D
Fri Nov 11 00:00:00 UTC 2022
MOXD1 knockdown suppresses the proliferation and tumor growth of glioblastoma cells via ER stress-inducing apoptosis.
Cell death discovery
Shi P,Xu J,Xia F,Wang Y,Ren J,Liang P,Cui H
Thu Apr 07 00:00:00 UTC 2022
L-Citrulline Supplementation Restrains Ferritinophagy-Mediated Ferroptosis to Alleviate Iron Overload-Induced Thymus Oxidative Damage and Immune Dysfunction.
Nutrients
Ba T,Zhao D,Chen Y,Zeng C,Zhang C,Niu S,Dai H
Fri Oct 28 00:00:00 UTC 2022
Hyaluronic Acid Oligosaccharide Derivatives Alleviate Lipopolysaccharide-Induced Inflammation in ATDC5 Cells by Multiple Mechanisms.
Molecules (Basel, Switzerland)
Huang H,Ding X,Xing D,Lin J,Li Z,Lin J
Wed Aug 31 00:00:00 UTC 2022
Hyaluronic Acid Oligosaccharide Derivatives Alleviate Lipopolysaccharide-Induced Inflammation in ATDC5 Cells by Multiple Mechanisms.
Molecules (Basel, Switzerland)
Huang H,Ding X,Xing D,Lin J,Li Z,Lin J
Wed Aug 31 00:00:00 UTC 2022
MiR-1298-5p level downregulation induced by Helicobacter pylori infection inhibits autophagy and promotes gastric cancer development by targeting MAP2K6.
Cellular signalling
Li X,Zhu M,Zhao G,Zhou A,Min L,Liu S,Zhang N,Zhu S,Guo Q,Zhang S,Li P
Sun May 01 00:00:00 UTC 2022
SIRT2 Affects Cell Proliferation and Apoptosis by Suppressing the Level of Autophagy in Renal Podocytes.
Disease markers
Liu S,Gao X,Fan Z,Wang Q
Tue May 03 00:00:00 UTC 2022
Moderate Treadmill Exercise Alleviates NAFLD by Regulating the Biogenesis and Autophagy of Lipid Droplet.
Nutrients
Yang Y,Li X,Liu Z,Ruan X,Wang H,Zhang Q,Cao L,Song L,Chen Y,Sun Y
Sun Nov 20 00:00:00 UTC 2022
Ferulic acid ameliorates renal injury via improving autophagy to inhibit inflammation in diabetic nephropathy mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Ma R,He Y,Fang Q,Xie G,Qi M
Thu Sep 01 00:00:00 UTC 2022
Ferulic acid ameliorates renal injury via improving autophagy to inhibit inflammation in diabetic nephropathy mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Ma R,He Y,Fang Q,Xie G,Qi M
Thu Sep 01 00:00:00 UTC 2022
Spermidine Ameliorates Nonalcoholic Steatohepatitis through Thyroid Hormone-Responsive Protein Signaling and the Gut Microbiota-Mediated Metabolism of Bile Acids.
Journal of agricultural and food chemistry
Ni Y,Hu Y,Lou X,Rong N,Liu F,Yang C,Zheng A,Yang S,Bao J,Fu Z
Wed Jun 01 00:00:00 UTC 2022
Spermidine Ameliorates Nonalcoholic Steatohepatitis through Thyroid Hormone-Responsive Protein Signaling and the Gut Microbiota-Mediated Metabolism of Bile Acids.
Journal of agricultural and food chemistry
Ni Y,Hu Y,Lou X,Rong N,Liu F,Yang C,Zheng A,Yang S,Bao J,Fu Z
Wed Jun 01 00:00:00 UTC 2022
Bcl-2 interacting protein 3 (BNIP3) promotes tumor growth in breast cancer under hypoxic conditions through an autophagy-dependent pathway.
Bioengineered
Zhang G,Xu Z,Yu M,Gao H
Tue Mar 01 00:00:00 UTC 2022
Astragalus-Scorpion Drug Pair Inhibits the Development of Prostate Cancer by Regulating GDPD4-2/PI3K/AKT/mTOR Pathway and Autophagy.
Frontiers in pharmacology
You X,Wu Y,Li Q,Sheng W,Zhou Q,Fu W
Tue Jul 19 00:00:00 UTC 2022
Astragalus-Scorpion Drug Pair Inhibits the Development of Prostate Cancer by Regulating GDPD4-2/PI3K/AKT/mTOR Pathway and Autophagy.
Frontiers in pharmacology
You X,Wu Y,Li Q,Sheng W,Zhou Q,Fu W
Tue Jul 19 00:00:00 UTC 2022
Biotinylated curcumin as a novel chemosensitizer enhances naphthalimide-induced autophagic cell death in breast cancer cells.
European journal of medicinal chemistry
Shao C,Wu J,Han S,Liu Y,Su Z,Zhu HL,Liu HK,Qian Y
Sat Jan 15 00:00:00 UTC 2022
Cathepsin B plays a role in spermatogenesis and sperm maturation through regulating autophagy and apoptosis in mice.
PeerJ
Wen Z,Zhu H,Wu B,Zhang A,Wang H,Cheng Y,Zhao H,Li J,Liu M,Gao J
Mon Jan 09 00:00:00 UTC 2023
Melatonin Attenuates Ropivacaine-Induced Apoptosis by Inhibiting Excessive Mitophagy Through the Parkin/PINK1 Pathway in PC12 and HT22 Cells.
Inflammation
Zeng L,He J,Liu C,Zhang F,Zhang Z,Chen H,Wang Q,Ding X,Luo H
Fri Apr 01 00:00:00 UTC 2022
Melatonin Attenuates Ropivacaine-Induced Apoptosis by Inhibiting Excessive Mitophagy Through the Parkin/PINK1 Pathway in PC12 and HT22 Cells.
Inflammation
Zeng L,He J,Liu C,Zhang F,Zhang Z,Chen H,Wang Q,Ding X,Luo H
Fri Apr 01 00:00:00 UTC 2022
Melatonin Attenuates Ropivacaine-Induced Apoptosis by Inhibiting Excessive Mitophagy Through the Parkin/PINK1 Pathway in PC12 and HT22 Cells.
Inflammation
Zeng L,He J,Liu C,Zhang F,Zhang Z,Chen H,Wang Q,Ding X,Luo H
Fri Apr 01 00:00:00 UTC 2022
Melatonin Attenuates Ropivacaine-Induced Apoptosis by Inhibiting Excessive Mitophagy Through the Parkin/PINK1 Pathway in PC12 and HT22 Cells.
Inflammation
Zeng L,He J,Liu C,Zhang F,Zhang Z,Chen H,Wang Q,Ding X,Luo H
Fri Apr 01 00:00:00 UTC 2022
PINK1-parkin-mediated neuronal mitophagy deficiency in prion disease.
Cell death & disease
Li J,Lai M,Zhang X,Li Z,Yang D,Zhao M,Wang D,Sun Z,Ehsan S,Li W,Gao H,Zhao D,Yang L
Fri Feb 18 00:00:00 UTC 2022
Autophagy and mitochondrial dynamics contribute to the protective effect of diosgenin against 3-MCPD induced kidney injury.
Chemico-biological interactions
Zhong Y,Sun D,Yao Y,Liu Q,Guo T,Wang X,Peng X
Tue Mar 01 00:00:00 UTC 2022
Mesenchymal stem cell-derived exosome mediated long non-coding RNA KLF3-AS1 represses autophagy and apoptosis of chondrocytes in osteoarthritis.
Cell cycle (Georgetown, Tex.)
Wen C,Lin L,Zou R,Lin F,Liu Y
Tue Feb 01 00:00:00 UTC 2022
Alliin alleviates LPS-induced pyroptosis via promoting mitophagy in THP-1 macrophages and mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Liu M,Lu J,Yang S,Chen Y,Yu J,Guan S
Tue Feb 01 00:00:00 UTC 2022
Exosomes-derived miR-125-5p from cartilage endplate stem cells regulates autophagy and ECM metabolism in nucleus pulposus by targeting SUV38H1.
Experimental cell research
Chen D,Jiang X
Sun May 01 00:00:00 UTC 2022
Exosomes-derived miR-125-5p from cartilage endplate stem cells regulates autophagy and ECM metabolism in nucleus pulposus by targeting SUV38H1.
Experimental cell research
Chen D,Jiang X
Sun May 01 00:00:00 UTC 2022
CDDO-Im ameliorates osteoarthritis and inhibits chondrocyte apoptosis in mice via enhancing Nrf2-dependent autophagy.
Acta pharmacologica Sinica
Dong J,Zhang KJ,Li GC,Chen XR,Lin JJ,Li JW,Lv ZY,Deng ZZ,Dai J,Cao W,Jiang Q
Fri Jul 01 00:00:00 UTC 2022
Genome-wide transcriptional profiling and functional analysis reveal miR-330-MAPK15 axis involving in cellular responses to deoxynivalenol exposure.
Chemosphere
Wang H,Zhou Y,Xu C,Cao Y,Xiao Y,Cai D,Uemoto Y,Wu S,Bao W
Fri Jul 01 00:00:00 UTC 2022
Autophagy Is a Crucial Path in Chondrogenesis of Adipose-Derived Mesenchymal Stromal Cells Laden in Hydrogel.
Gels (Basel, Switzerland)
Gabusi E,Lenzi E,Manferdini C,Dolzani P,Columbaro M,Saleh Y,Lisignoli G
Thu Nov 24 00:00:00 UTC 2022
Myo1b promotes tumor progression and angiogenesis by inhibiting autophagic degradation of HIF-1α in colorectal cancer.
Cell death & disease
Chen YH,Xu NZ,Hong C,Li WQ,Zhang YQ,Yu XY,Huang YL,Zhou JY
Tue Nov 08 00:00:00 UTC 2022
Dietary supplementation with metformin improves testis function and semen quality and increases antioxidants and autophagy capacity in goats.
Theriogenology
Zhao J,Yang PC,Yang H,Wang ZB,El-Samahy MA,Wang F,Zhang YL
Mon Aug 01 00:00:00 UTC 2022
S-20, a steroidal saponin from the berries of black nightshade, exerts anti-multidrug resistance activity in K562/ADR cells through autophagic cell death and ERK activation.
Food & function
Wang Y,Xu J,Wang Y,Xiang L,He X
Mon Feb 21 00:00:00 UTC 2022
Apelin-13/APJ induces cardiomyocyte hypertrophy by activating the Pannexin-1/P2X7 axis and FAM134B-dependent reticulophagy.
Journal of cellular physiology
Yang Y,Zhang K,Huang S,Chen W,Mao H,Ouyang X,Chen L,Li L
Fri Apr 01 00:00:00 UTC 2022
Apelin-13/APJ induces cardiomyocyte hypertrophy by activating the Pannexin-1/P2X7 axis and FAM134B-dependent reticulophagy.
Journal of cellular physiology
Yang Y,Zhang K,Huang S,Chen W,Mao H,Ouyang X,Chen L,Li L
Fri Apr 01 00:00:00 UTC 2022
Apelin-13/APJ induces cardiomyocyte hypertrophy by activating the Pannexin-1/P2X7 axis and FAM134B-dependent reticulophagy.
Journal of cellular physiology
Yang Y,Zhang K,Huang S,Chen W,Mao H,Ouyang X,Chen L,Li L
Fri Apr 01 00:00:00 UTC 2022
Apelin-13/APJ induces cardiomyocyte hypertrophy by activating the Pannexin-1/P2X7 axis and FAM134B-dependent reticulophagy.
Journal of cellular physiology
Yang Y,Zhang K,Huang S,Chen W,Mao H,Ouyang X,Chen L,Li L
Fri Apr 01 00:00:00 UTC 2022
Chelerythrine-Induced Apoptotic Cell Death in HepG2 Cells Involves the Inhibition of Akt Pathway and the Activation of Oxidative Stress and Mitochondrial Apoptotic Pathway.
Antioxidants (Basel, Switzerland)
Lin Y,Zhang Q,Xie B,Jiang H,Shen J,Tang S,Dai C
Sun Sep 18 00:00:00 UTC 2022
Levosimendan protects from sepsis-inducing cardiac dysfunction by suppressing inflammation, oxidative stress and regulating cardiac mitophagy via the PINK-1-Parkin pathway in mice.
Annals of translational medicine
Shi J,Chen Y,Zhi H,An H,Hu Z
Tue Feb 01 00:00:00 UTC 2022
Dl-3-n-butylphthalide attenuates myocardial ischemia reperfusion injury by suppressing oxidative stress and regulating cardiac mitophagy via the PINK1/Parkin pathway in rats.
Journal of thoracic disease
Zhang D,Zheng N,Fu X,Shi J,Zhang J
Sun May 01 00:00:00 UTC 2022
Dl-3-n-butylphthalide attenuates myocardial ischemia reperfusion injury by suppressing oxidative stress and regulating cardiac mitophagy via the PINK1/Parkin pathway in rats.
Journal of thoracic disease
Zhang D,Zheng N,Fu X,Shi J,Zhang J
Sun May 01 00:00:00 UTC 2022
RSL1D1 promotes the progression of colorectal cancer through RAN-mediated autophagy suppression.
Cell death & disease
Liu X,Chen J,Long X,Lan J,Liu X,Zhou M,Zhang S,Zhou J
Mon Jan 10 00:00:00 UTC 2022
RSL1D1 promotes the progression of colorectal cancer through RAN-mediated autophagy suppression.
Cell death & disease
Liu X,Chen J,Long X,Lan J,Liu X,Zhou M,Zhang S,Zhou J
Mon Jan 10 00:00:00 UTC 2022
Effects of butein on human osteosarcoma cell proliferation, apoptosis, and autophagy through oxidative stress.
Human & experimental toxicology
Zhang P,Zhang J,Quan H,Chen P,Wang J,Liang Y
Wed Mar 02 00:00:00 UTC 2022
Gynura segetum induces hepatic sinusoidal obstruction syndrome in mice by impairing autophagy.
Acta cirurgica brasileira
Zhang H,Jia S,Jin L,Mb JY,Shen Z,Wu J,Yao X,Chen D,Zhang C,Yu S,Zhu N,Jin L,Yao X
Thu Feb 24 00:00:00 UTC 2022
Pterostilbene attenuates intrauterine growth retardation-induced colon inflammation in piglets by modulating endoplasmic reticulum stress and autophagy.
Journal of animal science and biotechnology
Chen Y,Zhang H,Li Y,Ji S,Jia P,Wang T
Fri Nov 04 00:00:00 UTC 2022
Chicoric Acid Attenuated Renal Tubular Injury in HFD-Induced Chronic Kidney Disease Mice through the Promotion of Mitophagy via the Nrf2/PINK/Parkin Pathway.
Journal of agricultural and food chemistry
Ding XQ,Jian TY,Gai YN,Niu GT,Liu Y,Meng XH,Li J,Lyu H,Ren BR,Chen J
Wed Mar 09 00:00:00 UTC 2022
Chicoric Acid Attenuated Renal Tubular Injury in HFD-Induced Chronic Kidney Disease Mice through the Promotion of Mitophagy via the Nrf2/PINK/Parkin Pathway.
Journal of agricultural and food chemistry
Ding XQ,Jian TY,Gai YN,Niu GT,Liu Y,Meng XH,Li J,Lyu H,Ren BR,Chen J
Wed Mar 09 00:00:00 UTC 2022
Chicoric Acid Attenuated Renal Tubular Injury in HFD-Induced Chronic Kidney Disease Mice through the Promotion of Mitophagy via the Nrf2/PINK/Parkin Pathway.
Journal of agricultural and food chemistry
Ding XQ,Jian TY,Gai YN,Niu GT,Liu Y,Meng XH,Li J,Lyu H,Ren BR,Chen J
Wed Mar 09 00:00:00 UTC 2022
Chicoric Acid Attenuated Renal Tubular Injury in HFD-Induced Chronic Kidney Disease Mice through the Promotion of Mitophagy via the Nrf2/PINK/Parkin Pathway.
Journal of agricultural and food chemistry
Ding XQ,Jian TY,Gai YN,Niu GT,Liu Y,Meng XH,Li J,Lyu H,Ren BR,Chen J
Wed Mar 09 00:00:00 UTC 2022
Diallyl Trisulfide Enhances the Survival of Multiterritory Perforator Skin Flaps.
Frontiers in pharmacology
Dong C,Chen Z,Zhu L,Bsoul N,Wu H,Jiang J,Chen X,Lai Y,Yu G,Gu Y,Guo X,Gao W
Sat Mar 05 00:00:00 UTC 2022
Diallyl Trisulfide Enhances the Survival of Multiterritory Perforator Skin Flaps.
Frontiers in pharmacology
Dong C,Chen Z,Zhu L,Bsoul N,Wu H,Jiang J,Chen X,Lai Y,Yu G,Gu Y,Guo X,Gao W
Sat Mar 05 00:00:00 UTC 2022
Artemisinin Alleviates Cerebral Ischemia/Reperfusion-Induced Oxidative Damage via Regulating PHB2-Mediated Autophagy in the Human Neuroblastoma SH-SY5Y Cell Line.
Oxidative medicine and cellular longevity
Jiang M,Lai X,Zhang Y,Shen M,Ma H,Liu A,Wu J,Yan J
Tue Dec 27 00:00:00 UTC 2022
Artemisinin Alleviates Cerebral Ischemia/Reperfusion-Induced Oxidative Damage via Regulating PHB2-Mediated Autophagy in the Human Neuroblastoma SH-SY5Y Cell Line.
Oxidative medicine and cellular longevity
Jiang M,Lai X,Zhang Y,Shen M,Ma H,Liu A,Wu J,Yan J
Tue Dec 27 00:00:00 UTC 2022
Genome-wide CRISPR/Cas9 screen identifies host factors important for porcine reproductive and respiratory syndrome virus replication.
Virus research
Jiang J,Sun Y,Wang Y,Sabek A,Shangguan A,Wang K,Zhao S,Li G,Zhou A,Zhang S
Wed Jun 01 00:00:00 UTC 2022
Betulinic acid-nucleoside hybrid prevents acute alcohol -induced liver damage by promoting anti-oxidative stress and autophagy.
European journal of pharmacology
Zheng LY,Zou X,Wang YL,Zou M,Ma F,Wang N,Li JW,Wang MS,Hung HY,Wang Q
Wed Jan 05 00:00:00 UTC 2022
Betulinic acid-nucleoside hybrid prevents acute alcohol -induced liver damage by promoting anti-oxidative stress and autophagy.
European journal of pharmacology
Zheng LY,Zou X,Wang YL,Zou M,Ma F,Wang N,Li JW,Wang MS,Hung HY,Wang Q
Wed Jan 05 00:00:00 UTC 2022
Cold exposure-induced endoplasmic reticulum stress regulates autophagy through the SIRT2/FoxO1 signaling pathway.
Journal of cellular physiology
Guo J,Nie J,Chen Z,Wang X,Hu H,Xu J,Lu J,Ma L,Ji H,Yuan J,Xu B
Sat Oct 01 00:00:00 UTC 2022
Epimedokoreanin B inhibits the growth of lung cancer cells through endoplasmic reticulum stress-mediated paraptosis accompanied by autophagosome accumulation.
Chemico-biological interactions
Zheng H,Liu Q,Wang S,Liu X,Ma M,Shen T,Wang X,Ren D
Sat Oct 01 00:00:00 UTC 2022
ZC3H4 regulates infiltrating monocytes, attenuating pulmonary fibrosis through IL-10.
Respiratory research
Liu Y,Zhang X,Wang J,Yang F,Luo W,Huang J,Chen M,Wang S,Li C,Zhang W,Chao J
Fri Aug 12 00:00:00 UTC 2022
β-Carotene Attenuates Apoptosis and Autophagy via PI3K/AKT/mTOR Signaling Pathway in Necrotizing Enterocolitis Model Cells IEC-6.
Evidence-based complementary and alternative medicine : eCAM
Xu G,Ma T,Zhou C,Zhao F,Peng K,Li B
Sat Jul 16 00:00:00 UTC 2022
Arsenic Prodrug-Mediated Tumor Microenvironment Modulation Platform for Synergetic Glioblastoma Therapy.
ACS applied materials & interfaces
Yan J,Hanif S,Zhang D,Ismail M,Wang X,Li Q,Shi B,Muhammad P,Wu H
Wed Aug 17 00:00:00 UTC 2022
Dysregulated autophagy-related genes in septic cardiomyopathy: Comprehensive bioinformatics analysis based on the human transcriptomes and experimental validation.
Frontiers in cardiovascular medicine
Zou HX,Qiu BQ,Zhang ZY,Hu T,Wan L,Liu JC,Huang H,Lai SQ
Sat Aug 20 00:00:00 UTC 2022
Dysregulated autophagy-related genes in septic cardiomyopathy: Comprehensive bioinformatics analysis based on the human transcriptomes and experimental validation.
Frontiers in cardiovascular medicine
Zou HX,Qiu BQ,Zhang ZY,Hu T,Wan L,Liu JC,Huang H,Lai SQ
Sat Aug 20 00:00:00 UTC 2022
Inhibition of STAT3-ferroptosis negative regulatory axis suppresses tumor growth and alleviates chemoresistance in gastric cancer.
Redox biology
Ouyang S,Li H,Lou L,Huang Q,Zhang Z,Mo J,Li M,Lu J,Zhu K,Chu Y,Ding W,Zhu J,Lin Z,Zhong L,Wang J,Yue P,Turkson J,Liu P,Wang Y,Zhang X
Wed Jun 01 00:00:00 UTC 2022
Metformin promotes histone deacetylation of optineurin and suppresses tumour growth through autophagy inhibition in ocular melanoma.
Clinical and translational medicine
Zhuang A,Chai P,Wang S,Zuo S,Yu J,Jia S,Ge S,Jia R,Zhou Y,Shi W,Xu X,Ruan J,Fan X
Sat Jan 01 00:00:00 UTC 2022
LRPPRC inhibits autophagy and promotes foam cell formation in atherosclerosis.
The FEBS journal
Xu Z,Li X,Ding Z,Zhang Y,Peng Z,Yang X,Cao W,Du R
Thu Dec 01 00:00:00 UTC 2022
LRPPRC inhibits autophagy and promotes foam cell formation in atherosclerosis.
The FEBS journal
Xu Z,Li X,Ding Z,Zhang Y,Peng Z,Yang X,Cao W,Du R
Thu Dec 01 00:00:00 UTC 2022
Celastrol upregulated ATG7 triggers autophagy via targeting Nur77 in colorectal cancer.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Zhang W,Wu Z,Qi H,Chen L,Wang T,Mao X,Shi H,Chen H,Zhong M,Shi X,Wang X,Li Q
Thu Sep 01 00:00:00 UTC 2022
IL-17A promotes lung fibrosis through impairing mitochondrial homeostasis in type II alveolar epithelial cells.
Journal of cellular and molecular medicine
Xiao H,Peng L,Jiang D,Liu Y,Zhu L,Li Z,Geng J,Xie B,Huang X,Wang J,Dai H,Wang C
Tue Nov 01 00:00:00 UTC 2022
Extracellular vesicle miR-32 derived from macrophage promotes arterial calcification in mice with type 2 diabetes via inhibiting VSMC autophagy.
Journal of translational medicine
Cao J,Chen C,Chen Q,Gao Y,Zhao Z,Yuan Q,Li A,Yang S,He Y,Zu X,Liu J
Wed Jul 06 00:00:00 UTC 2022
Resveratrol Synergistically Promotes BMP9-Induced Osteogenic Differentiation of Mesenchymal Stem Cells.
Stem cells international
Wang Y,Xia C,Chen Y,Jiang T,Hu Y,Gao Y
Fri Aug 05 00:00:00 UTC 2022
Astaxanthin Activated the Nrf2/HO-1 Pathway to Enhance Autophagy and Inhibit Ferroptosis, Ameliorating Acetaminophen-Induced Liver Injury.
ACS applied materials & interfaces
Cai X,Hua S,Deng J,Du Z,Zhang D,Liu Z,Khan NU,Zhou M,Chen Z
Wed Sep 28 00:00:00 UTC 2022
Podophyllotoxin-combined 5-aminolevulinic acid photodynamic therapy significantly promotes HR-HPV-infected cell death.
Photodermatology, photoimmunology & photomedicine
Wang J,Wang Q,Chen P,Li Q,Li Z,Xu M,Zeng K,Li C
Fri Jul 01 00:00:00 UTC 2022
Podophyllotoxin-combined 5-aminolevulinic acid photodynamic therapy significantly promotes HR-HPV-infected cell death.
Photodermatology, photoimmunology & photomedicine
Wang J,Wang Q,Chen P,Li Q,Li Z,Xu M,Zeng K,Li C
Fri Jul 01 00:00:00 UTC 2022
Gold nanoparticles targeting the autophagy-lysosome system to combat the inflammation-compromised osteogenic potential of periodontal ligament stem cells: From mechanism to therapy.
Biomaterials
Yin Y,Tian BM,Li X,Yu YC,Deng DK,Sun LJ,Qu HL,Wu RX,Xu XY,Sun HH,An Y,He XT,Chen FM
Thu Sep 01 00:00:00 UTC 2022
Gold nanoparticles targeting the autophagy-lysosome system to combat the inflammation-compromised osteogenic potential of periodontal ligament stem cells: From mechanism to therapy.
Biomaterials
Yin Y,Tian BM,Li X,Yu YC,Deng DK,Sun LJ,Qu HL,Wu RX,Xu XY,Sun HH,An Y,He XT,Chen FM
Thu Sep 01 00:00:00 UTC 2022
Lactoferrin alleviates spermatogenesis dysfunction caused by bisphenol A and cadmium via ameliorating disordered autophagy, apoptosis and oxidative stress.
International journal of biological macromolecules
He H,Chen X,Li X,Yang K,Li J,Shi H
Thu Dec 01 00:00:00 UTC 2022
Parkin-independent mitophagy via Drp1-mediated outer membrane severing and inner membrane ubiquitination.
The Journal of cell biology
Oshima Y,Cartier E,Boyman L,Verhoeven N,Polster BM,Huang W,Kane M,Lederer WJ,Karbowski M
Mon Jun 07 00:00:00 UTC 2021
Parkin-independent mitophagy via Drp1-mediated outer membrane severing and inner membrane ubiquitination.
The Journal of cell biology
Oshima Y,Cartier E,Boyman L,Verhoeven N,Polster BM,Huang W,Kane M,Lederer WJ,Karbowski M
Mon Jun 07 00:00:00 UTC 2021
PPARγ/NF-κB and TGF-β1/Smad pathway are involved in the anti-fibrotic effects of levo-tetrahydropalmatine on liver fibrosis.
Journal of cellular and molecular medicine
Yu Q,Cheng P,Wu J,Guo C
Mon Feb 01 00:00:00 UTC 2021
PPARγ/NF-κB and TGF-β1/Smad pathway are involved in the anti-fibrotic effects of levo-tetrahydropalmatine on liver fibrosis.
Journal of cellular and molecular medicine
Yu Q,Cheng P,Wu J,Guo C
Mon Feb 01 00:00:00 UTC 2021
PPARγ/NF-κB and TGF-β1/Smad pathway are involved in the anti-fibrotic effects of levo-tetrahydropalmatine on liver fibrosis.
Journal of cellular and molecular medicine
Yu Q,Cheng P,Wu J,Guo C
Mon Feb 01 00:00:00 UTC 2021
Knockdown of VDAC1 alleviates the cognitive dysfunction secondary to sepsis-associated encephalopathy.
American journal of translational research
Cai M,Du B,Si Y,Miao J,Ge J,Zhang J,Song J,Bao H
Thu Nov 18 00:00:00 UTC 2021
FAM134B-Mediated ER-phagy Upregulation Attenuates AGEs-Induced Apoptosis and Senescence in Human Nucleus Pulposus Cells.
Oxidative medicine and cellular longevity
Luo R,Li S,Li G,Lu S,Zhang W,Liu H,Lei J,Ma L,Ke W,Liao Z,Wang B,Song Y,Wang K,Zhang Y,Yang C
Wed Jan 05 00:00:00 UTC 2022
Fenofibrate Ameliorates Hepatic Ischemia/Reperfusion Injury in Mice: Involvements of Apoptosis, Autophagy, and PPAR-α Activation.
PPAR research
Zhang J,Cheng P,Dai W,Ji J,Wu L,Feng J,Wu J,Yu Q,Li J,Guo C
Sat Feb 20 00:00:00 UTC 2021
Procyanidin B2 alleviates liver injury caused by cold stimulation through Sonic hedgehog signalling and autophagy.
Journal of cellular and molecular medicine
Ma L,Li C,Lian S,Xu B,Lv H,Liu Y,Lu J,Ji H,Li S,Guo J,Yang H
Sun Aug 01 00:00:00 UTC 2021
Reduction of Autophagosome Overload Attenuates Neuronal Cell Death After Traumatic Brain Injury.
Neuroscience
Quan X,Song L,Zheng X,Liu S,Ding H,Li S,Xu G,Li X,Liu L
Thu Apr 15 00:00:00 UTC 2021
Aiduqing formula suppresses breast cancer metastasis via inhibiting CXCL1-mediated autophagy.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yang B,Peng F,Zhang Y,Wang X,Wang S,Zheng Y,Zhang J,Zeng Y,Wang N,Peng C,Wang Z
Wed Sep 01 00:00:00 UTC 2021
Aiduqing formula suppresses breast cancer metastasis via inhibiting CXCL1-mediated autophagy.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yang B,Peng F,Zhang Y,Wang X,Wang S,Zheng Y,Zhang J,Zeng Y,Wang N,Peng C,Wang Z
Wed Sep 01 00:00:00 UTC 2021
A-24, a steroidal saponin from Allium chinense, induced apoptosis, autophagy and migration inhibition in p53 wild-type and p53-deficient gastric cancer cells.
Chemico-biological interactions
Xu J,Wang Y,Wang Y,Wang Z,He X
Fri Oct 01 00:00:00 UTC 2021
Advanced oxidation protein products impair autophagic flux in macrophage by inducing lysosomal dysfunction via activation of PI3K-Akt-mTOR pathway in Crohn's disease.
Free radical biology & medicine
Liao Y,Xu J,Qin B,Shi J,Qin C,Xie F,Ou S,Tang J,Wang W,Wu F,Bai L
Fri Aug 20 00:00:00 UTC 2021
Advanced oxidation protein products impair autophagic flux in macrophage by inducing lysosomal dysfunction via activation of PI3K-Akt-mTOR pathway in Crohn's disease.
Free radical biology & medicine
Liao Y,Xu J,Qin B,Shi J,Qin C,Xie F,Ou S,Tang J,Wang W,Wu F,Bai L
Fri Aug 20 00:00:00 UTC 2021
Study on the anti-hepatocarcinoma effect and molecular mechanism of Prunella vulgaris total flavonoids.
Journal of ethnopharmacology
Song YG,Kang L,Tian S,Cui LL,Li Y,Bai M,Fang XY,Cao LH,Coleman K,Miao MS
Sat Jun 12 00:00:00 UTC 2021
Study on the anti-hepatocarcinoma effect and molecular mechanism of Prunella vulgaris total flavonoids.
Journal of ethnopharmacology
Song YG,Kang L,Tian S,Cui LL,Li Y,Bai M,Fang XY,Cao LH,Coleman K,Miao MS
Sat Jun 12 00:00:00 UTC 2021
T-17, a spirostanol saponin, inhibits p53-independent proliferation and p53-dependent migration of gastric cancer cells.
Steroids
Xu J,Wang Y,Wang Z,Wang Y,He X
Tue Jun 01 00:00:00 UTC 2021
WIPI-1 inhibits metastasis and tumour growth via the WIPI-1-TRIM21 axis and MYC regulation in nasopharyngeal carcinoma.
Oral oncology
Zhao Y,Li WF,Li QJ,He SW,He QM,Long LF,Liu N,Ma J
Mon Nov 01 00:00:00 UTC 2021
Cannabidiol inhibits human glioma by induction of lethal mitophagy through activating TRPV4.
Autophagy
Huang T,Xu T,Wang Y,Zhou Y,Yu D,Wang Z,He L,Chen Z,Zhang Y,Davidson D,Dai Y,Hang C,Liu X,Yan C
Mon Nov 01 00:00:00 UTC 2021
Cannabidiol inhibits human glioma by induction of lethal mitophagy through activating TRPV4.
Autophagy
Huang T,Xu T,Wang Y,Zhou Y,Yu D,Wang Z,He L,Chen Z,Zhang Y,Davidson D,Dai Y,Hang C,Liu X,Yan C
Mon Nov 01 00:00:00 UTC 2021
Cannabidiol inhibits human glioma by induction of lethal mitophagy through activating TRPV4.
Autophagy
Huang T,Xu T,Wang Y,Zhou Y,Yu D,Wang Z,He L,Chen Z,Zhang Y,Davidson D,Dai Y,Hang C,Liu X,Yan C
Mon Nov 01 00:00:00 UTC 2021
Magnesium-Assisted Cisplatin Inhibits Bladder Cancer Cell Survival by Modulating Wnt/β-Catenin Signaling Pathway.
Frontiers in pharmacology
Li T,Tang Z,Li C,Liu X,Cheng L,Yang Z,Zhu X,Liu W,Huang Y
Wed Feb 16 00:00:00 UTC 2022
Astragaloside IV alleviates liver injury in type 2 diabetes due to promotion of AMPK/mTOR‑mediated autophagy.
Molecular medicine reports
Zhu Y,Su Y,Zhang J,Zhang Y,Li Y,Han Y,Dong X,Li W,Li W
Tue Jun 01 00:00:00 UTC 2021
Sorafenib-Induced Autophagy Promotes Glycolysis by Upregulating the p62/HDAC6/HSP90 Axis in Hepatocellular Carcinoma Cells.
Frontiers in pharmacology
Yan X,Tian R,Sun J,Zhao Y,Liu B,Su J,Li M,Sun W,Xu X
Tue Mar 08 00:00:00 UTC 2022
CHNQD-00603 Promotes Osteogenic Differentiation of Bone Marrow Mesenchymal Stem Cells by the miR-452-3p-Mediated Autophagy Pathway.
Frontiers in cell and developmental biology
Xin S,Li SM,Gao L,Zheng JJ,Wu YW,Shao CL,Ren WH,Zhi K
Sat Jan 08 00:00:00 UTC 2022
Autophagy is required for the promoting effect of angiogenic factor with G patch domain and forkhead-associated domain 1 (AGGF1) in retinal angiogenesis.
Microvascular research
Li R,Yao G,Zhou L,Zhang M,Yan J,Wang X,Li Y
Mon Nov 01 00:00:00 UTC 2021
Chorionic villus-derived mesenchymal stem cell-mediated autophagy promotes the proliferation and invasiveness of trophoblasts under hypoxia by activating the JAK2/STAT3 signalling pathway.
Cell & bioscience
Chu Y,Zhu C,Yue C,Peng W,Chen W,He G,Liu C,Lv Y,Gao G,Yao K,Han R,Hu X,Zhang Y,Ye Y
Wed Oct 13 00:00:00 UTC 2021
Metformin attenuates atherosclerosis and plaque vulnerability by upregulating KLF2-mediated autophagy in apoE-/- mice.
Biochemical and biophysical research communications
Wu H,Feng K,Zhang C,Zhang H,Zhang J,Hua Y,Dong Z,Zhu Y,Yang S,Ma C
Fri Jun 11 00:00:00 UTC 2021
Metformin attenuates atherosclerosis and plaque vulnerability by upregulating KLF2-mediated autophagy in apoE-/- mice.
Biochemical and biophysical research communications
Wu H,Feng K,Zhang C,Zhang H,Zhang J,Hua Y,Dong Z,Zhu Y,Yang S,Ma C
Fri Jun 11 00:00:00 UTC 2021
Gastrodin ameliorates learning and memory impairment in rats with vascular dementia by promoting autophagy flux via inhibition of the Ca2+/CaMKII signal pathway.
Aging
Chen TT,Zhou X,Xu YN,Li Y,Wu XY,Xiang Q,Fu LY,Hu XX,Tao L,Shen XC
Wed Mar 10 00:00:00 UTC 2021
Exposure to DEHP induces testis toxicity and injury through the ROS/mTOR/NLRP3 signaling pathway in immature rats.
Ecotoxicology and environmental safety
Hong Y,Zhou Y,Shen L,Wei Y,Long C,Fu Y,Wu H,Wang J,Wu Y,Wu S,Wei G
Mon Dec 20 00:00:00 UTC 2021
Exposure to DEHP induces testis toxicity and injury through the ROS/mTOR/NLRP3 signaling pathway in immature rats.
Ecotoxicology and environmental safety
Hong Y,Zhou Y,Shen L,Wei Y,Long C,Fu Y,Wu H,Wang J,Wu Y,Wu S,Wei G
Mon Dec 20 00:00:00 UTC 2021
Rosuvastatin exerts anti-atherosclerotic effects by improving macrophage-related foam cell formation and polarization conversion via mediating autophagic activities.
Journal of translational medicine
Zhang X,Qin Y,Wan X,Liu H,Lv C,Ruan W,He L,Lu L,Guo X
Wed Feb 10 00:00:00 UTC 2021
DJ-1 Deficiency in Hepatocytes Improves Liver Ischemia-Reperfusion Injury by Enhancing Mitophagy.
Cellular and molecular gastroenterology and hepatology
Xu M,Hang H,Huang M,Li J,Xu D,Jiao J,Wang F,Wu H,Sun X,Gu J,Kong X,Gao Y
Thu Jan 27 00:00:00 UTC 2022
Astragaloside II Ameliorated Podocyte Injury and Mitochondrial Dysfunction in Streptozotocin-Induced Diabetic Rats.
Frontiers in pharmacology
Su J,Gao C,Xie L,Fan Y,Shen Y,Huang Q,Wang N,Xu Y,Yang N,Gui D
Sat Apr 03 00:00:00 UTC 2021
Autophagy is Involved in Neuroprotective Effect of Alpha7 Nicotinic Acetylcholine Receptor on Ischemic Stroke.
Frontiers in pharmacology
Xu ZQ,Zhang JJ,Kong N,Zhang GY,Ke P,Han T,Su DF,Liu C
Tue May 18 00:00:00 UTC 2021
Autophagy is Involved in Neuroprotective Effect of Alpha7 Nicotinic Acetylcholine Receptor on Ischemic Stroke.
Frontiers in pharmacology
Xu ZQ,Zhang JJ,Kong N,Zhang GY,Ke P,Han T,Su DF,Liu C
Tue May 18 00:00:00 UTC 2021
Silencing circular RNA circ_0054537 and upregulating microRNA-640 suppress malignant progression of renal cell carcinoma via regulating neuronal pentraxin-2 (NPTX2).
Bioengineered
Pei L,Lv X,Jia G,Tan X,Li M,Zhang A
Wed Dec 01 00:00:00 UTC 2021
N-linoleyltyrosine exerts neuroprotective effects in APP/PS1 transgenic mice via cannabinoid receptor-mediated autophagy.
Journal of pharmacological sciences
Long CM,Zheng QX,Zhou Y,Liu YT,Gong LP,Zeng YC,Liu S
Wed Dec 01 00:00:00 UTC 2021
Melatonin ameliorates cerebral ischemia-reperfusion injury in diabetic mice by enhancing autophagy via the SIRT1-BMAL1 pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Liu L,Cao Q,Gao W,Li BY,Zeng C,Xia Z,Zhao B
Wed Dec 01 00:00:00 UTC 2021
Circadian misalignment promotes vascular smooth muscle cell apoptosis via defective autophagy.
Journal of molecular histology
Guo Z,Yu B,Li X,Yang X,Wang C,Fan L
Sun Aug 01 00:00:00 UTC 2021
Interplay between MTOR and GPX4 signaling modulates autophagy-dependent ferroptotic cancer cell death.
Cancer gene therapy
Liu Y,Wang Y,Liu J,Kang R,Tang D
Mon Feb 01 00:00:00 UTC 2021
Resveratrol attenuates cigarette smoke induced endothelial apoptosis by activating Notch1 signaling mediated autophagy.
Respiratory research
Zong DD,Liu XM,Li JH,Ouyang RY,Long YJ,Chen P,Chen Y
Tue Jan 19 00:00:00 UTC 2021
Apatinib triggers autophagic and apoptotic cell death via VEGFR2/STAT3/PD-L1 and ROS/Nrf2/p62 signaling in lung cancer.
Journal of experimental & clinical cancer research : CR
Xie C,Zhou X,Liang C,Li X,Ge M,Chen Y,Yin J,Zhu J,Zhong C
Tue Aug 24 00:00:00 UTC 2021
Apatinib triggers autophagic and apoptotic cell death via VEGFR2/STAT3/PD-L1 and ROS/Nrf2/p62 signaling in lung cancer.
Journal of experimental & clinical cancer research : CR
Xie C,Zhou X,Liang C,Li X,Ge M,Chen Y,Yin J,Zhu J,Zhong C
Tue Aug 24 00:00:00 UTC 2021
Apatinib triggers autophagic and apoptotic cell death via VEGFR2/STAT3/PD-L1 and ROS/Nrf2/p62 signaling in lung cancer.
Journal of experimental & clinical cancer research : CR
Xie C,Zhou X,Liang C,Li X,Ge M,Chen Y,Yin J,Zhu J,Zhong C
Tue Aug 24 00:00:00 UTC 2021
Decreased neuronal synaptosome associated protein 29 contributes to poststroke cognitive impairment by disrupting presynaptic maintenance.
Theranostics
Yan W,Fan J,Zhang X,Song H,Wan R,Wang W,Yin Y
Mon Aug 16 00:00:00 UTC 2021
RabGEF1 functions as an oncogene in U251 glioblastoma cells and is involved in regulating AKT and Erk pathways.
Experimental and molecular pathology
Fan H,Xin T,Dong X,Yang F,Zhang R,Feng S,He D,Guo H,Pang Q
Mon Feb 01 00:00:00 UTC 2021
1,3-dichloro-2-propanol induced hepatic lipid accumulation by inhibiting autophagy via AKT/mTOR/FOXO1 pathway in mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fan Y,Lu J,Liu J,Zhang R,Yu Z,Guan S
Mon Nov 01 00:00:00 UTC 2021
A Marine λ-Oligocarrageenan Inhibits Migratory and Invasive Ability of MDA-MB-231 Human Breast Cancer Cells through Actions on Heparanase Metabolism and MMP-14/MMP-2 Axis.
Marine drugs
Cousin R,Groult H,Manseur C,Ferru-Clément R,Gani M,Havret R,Toucheteau C,Prunier G,Colin B,Morel F,Piot JM,Lanneluc I,Baranger K,Maugard T,Fruitier-Arnaudin I
Tue Sep 28 00:00:00 UTC 2021
miR-338-3p inhibits autophagy in a rat model of allergic rhinitis after PM2.5 exposure through AKT/mTOR signaling by targeting UBE2Q1.
Biochemical and biophysical research communications
Wang JC,Huang Y,Zhang RX,Han ZJ,Zhou LL,Sun N,Dong WY,Zhuang GS
Fri May 21 00:00:00 UTC 2021
The anticancer activity of root extract from Quercus acutissima Carruth. Via regulating apoptosis and autophagy in breast cancer cells.
Journal of ethnopharmacology
Chen C,Ding W,Li J,Geng X,Zhang H,Sun Y
Fri Oct 08 00:00:00 UTC 2021
Co-modification with MSC membrane and PDA prevents Fe3O4-induced pulmonary toxicity in mice via AMPK-ULK1 axis.
Toxicology letters
Yao H,Yan J,Shao P,Wang Y,Liu T,Jiang J,Liu T
Fri Oct 15 00:00:00 UTC 2021
Dl-3-n-Butylphthalide Reduces Cognitive Deficits and Alleviates Neuropathology in P301S Tau Transgenic Mice.
Frontiers in neuroscience
Chang Y,Yao Y,Ma R,Wang Z,Hu J,Wu Y,Jiang X,Li L,Li G
Mon Jul 12 00:00:00 UTC 2021
EphA2 inhibits SRA01/04 cells apoptosis by suppressing autophagy via activating PI3K/Akt/mTOR pathway.
Archives of biochemistry and biophysics
Han J,Wang L,Lv H,Liu J,Dong Y,Shi L,Ji Q
Sat Oct 30 00:00:00 UTC 2021
OM-MSCs Alleviate the Golgi Apparatus Stress Response following Cerebral Ischemia/Reperfusion Injury via the PEDF-PI3K/Akt/mTOR Signaling Pathway.
Oxidative medicine and cellular longevity
He J,Liu J,Huang Y,Tang X,Xiao H,Liu Z,Jiang Z,Zeng L,Hu Z,Lu M
Mon Feb 07 00:00:00 UTC 2022
Macroautophagy supports Sonic Hedgehog signaling by promoting Patched1 degradation.
Biochimica et biophysica acta. Molecular cell research
Yang X,Jin N,Wang Y,Yao Y,Wang Y,Li T,Liu C,Yu T,Yin H,Zhang Z,Cheng SY,Yue S
Mon Nov 01 00:00:00 UTC 2021
Icariin improves brain function decline in aging rats by enhancing neuronal autophagy through the AMPK/mTOR/ULK1 pathway.
Pharmaceutical biology
Zheng J,Hu S,Wang J,Zhang X,Yuan D,Zhang C,Liu C,Wang T,Zhou Z
Wed Dec 01 00:00:00 UTC 2021
Blockage of Extracellular Signal-Regulated Kinase Exerts an Antitumor Effect via Regulating Energy Metabolism and Enhances the Efficacy of Autophagy Inhibitors by Regulating Transcription Factor EB Nuclear Translocation in Osteosarcoma.
Frontiers in cell and developmental biology
Zhang M,Bai Y,Xu C,Qi Y,Meng J,Zhang W,Su H,Yan W
Sat Aug 21 00:00:00 UTC 2021
PFKP facilitates ATG4B phosphorylation during amino acid deprivation-induced autophagy.
Cellular signalling
Li X,Sun L,Yan G,Yan X
Tue Jun 01 00:00:00 UTC 2021
Atorvastatin reverses high cholesterol-induced cardiac remodelling and regulates mitochondrial quality-control in a cholesterol-independent manner: An experimental study.
Clinical and experimental pharmacology & physiology
Zheng P,Ding Y,Lu F,Liu N,Wu H,Bian Z,Chen X,Yang D
Sun Aug 01 00:00:00 UTC 2021
Atorvastatin reverses high cholesterol-induced cardiac remodelling and regulates mitochondrial quality-control in a cholesterol-independent manner: An experimental study.
Clinical and experimental pharmacology & physiology
Zheng P,Ding Y,Lu F,Liu N,Wu H,Bian Z,Chen X,Yang D
Sun Aug 01 00:00:00 UTC 2021
Bone Autophagy: A Potential Way of Exercise-Mediated Meg3/P62/Runx2 Pathway to Regulate Bone Formation in T2DM Mice.
Diabetes, metabolic syndrome and obesity : targets and therapy
Chen X,Yang K,Jin X,Meng Z,Liu B,Yu H,Lu P,Wang K,Fan Z,Tang Z,Zhang F,Liu C
Sun Apr 24 00:00:00 UTC 2022
Aflatoxin B1 inhibited autophagy flux by inducing lysosomal alkalinization in HepG2 cells.
Toxicology mechanisms and methods
Zhang Q,Feng Z,Lu J,Lu J,Guan S,Chen Y
Thu Jul 01 00:00:00 UTC 2021
Hsa_circ_0092276 promotes doxorubicin resistance in breast cancer cells by regulating autophagy via miR-348/ATG7 axis.
Translational oncology
Wang Q,Liang D,Shen P,Yu Y,Yan Y,You W
Sun Aug 01 00:00:00 UTC 2021
An integrative transcriptome study reveals Ddit4/Redd1 as a key regulator of cancer cachexia in rodent models.
Cell death & disease
Niu M,Li L,Su Z,Wei L,Pu W,Zhao C,Ding Y,Wazir J,Cao W,Song S,Gao Q,Wang H
Sat Jun 26 00:00:00 UTC 2021
SNHG9, a Papillary Thyroid Cancer Cell Exosome-Enriched lncRNA, Inhibits Cell Autophagy and Promotes Cell Apoptosis of Normal Thyroid Epithelial Cell Nthy-ori-3 Through YBOX3/P21 Pathway.
Frontiers in oncology
Wen D,Liu WL,Lu ZW,Cao YM,Ji QH,Wei WJ
Sun May 23 00:00:00 UTC 2021
Luteolin Alleviates Methamphetamine-Induced Hepatotoxicity by Suppressing the p53 Pathway-Mediated Apoptosis, Autophagy, and Inflammation in Rats.
Frontiers in pharmacology
Zhang KK,Wang H,Qu D,Chen LJ,Wang LB,Li JH,Liu JL,Xu LL,Yoshida JS,Xu JT,Xie XL,Li DR
Wed Mar 10 00:00:00 UTC 2021
Long non-coding RNA LINC01559 exerts oncogenic role via enhancing autophagy in lung adenocarcinoma.
Cancer cell international
Zhao Z,Wan J,Guo M,Yang Z,Li Z,Wang Y,Ming L
Thu Nov 25 00:00:00 UTC 2021
Long non-coding RNA LINC01559 exerts oncogenic role via enhancing autophagy in lung adenocarcinoma.
Cancer cell international
Zhao Z,Wan J,Guo M,Yang Z,Li Z,Wang Y,Ming L
Thu Nov 25 00:00:00 UTC 2021
Salidroside alleviates taurolithocholic acid 3-sulfate-induced AR42J cell injury.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Qian J,Wang X,Weng W,Zhou G,Zhu S,Liu C
Fri Oct 01 00:00:00 UTC 2021
2-Amino-3-Methylimidazo[4,5-f]quinoline Triggering Liver Damage by Inhibiting Autophagy and Inducing Endoplasmic Reticulum Stress in Zebrafish (Danio rerio).
Toxins
Li D,Li Z,Zhang T,Peng B,Zhang Y,Sun H,Wang S
Mon Nov 22 00:00:00 UTC 2021
Rab26 suppresses migration and invasion of breast cancer cells through mediating autophagic degradation of phosphorylated Src.
Cell death & disease
Liu H,Zhou Y,Qiu H,Zhuang R,Han Y,Liu X,Qiu X,Wang Z,Xu L,Tan R,Hong W,Wang T
Wed Mar 17 00:00:00 UTC 2021
The enhancement of Tetrandrine to gemcitabine-resistant PANC-1 cytochemical sensitivity involves the promotion of PI3K/Akt/mTOR-mediated apoptosis and AMPK-regulated autophagy.
Acta histochemica
Song J,Xu J,Guo J,Shang Y,Wang J,Wang T
Wed Sep 01 00:00:00 UTC 2021
Nlp promotes autophagy through facilitating the interaction of Rab7 and FYCO1.
Signal transduction and targeted therapy
Xiao W,Yeerken D,Li J,Li Z,Jiang L,Li D,Fu M,Ma L,Song Y,Zhang W,Zhan Q
Fri Apr 16 00:00:00 UTC 2021
Nlp promotes autophagy through facilitating the interaction of Rab7 and FYCO1.
Signal transduction and targeted therapy
Xiao W,Yeerken D,Li J,Li Z,Jiang L,Li D,Fu M,Ma L,Song Y,Zhang W,Zhan Q
Fri Apr 16 00:00:00 UTC 2021
Berberine Inhibits MDA-MB-231 Cells as an Agonist of G Protein-Coupled Estrogen Receptor 1.
International journal of molecular sciences
Qi M,Liu X,Zhou Y,Wang H,Zhao Y,Ren J,Xiang J
Sun Oct 24 00:00:00 UTC 2021
17β-estradiol improves the developmental ability, inhibits reactive oxygen species levels and apoptosis of porcine oocytes by regulating autophagy events.
The Journal of steroid biochemistry and molecular biology
Duan J,Chen H,Xu D,Li Y,Li X,Cheng J,Hua R,Zhang Z,Yang L,Li Q
Sat May 01 00:00:00 UTC 2021
LncRNA FEZF1-AS1 Promotes Multi-Drug Resistance of Gastric Cancer Cells via Upregulating ATG5.
Frontiers in cell and developmental biology
Gui Z,Zhao Z,Sun Q,Shao G,Huang J,Zhao W,Kuang Y
Fri Nov 19 00:00:00 UTC 2021
Lanthanum chloride induces autophagy in primary cultured rat cortical neurons through Akt/mTOR and AMPK/mTOR signaling pathways.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Gao X,Yu M,Sun W,Han Y,Yang J,Lu X,Jin C,Wu S,Cai Y
Wed Dec 01 00:00:00 UTC 2021
Lanthanum chloride induces autophagy in primary cultured rat cortical neurons through Akt/mTOR and AMPK/mTOR signaling pathways.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Gao X,Yu M,Sun W,Han Y,Yang J,Lu X,Jin C,Wu S,Cai Y
Wed Dec 01 00:00:00 UTC 2021
CIDEC silencing attenuates diabetic nephropathy via inhibiting apoptosis and promoting autophagy.
Journal of diabetes investigation
Zheng GS,Tan YM,Shang YY,Liu YP,Hu BA,Wang D,Han L,Wang ZH,Zhang W,Ti Y,Zhong M
Sun Aug 01 00:00:00 UTC 2021
Circ-SIRT1 inhibits cardiac hypertrophy via activating SIRT1 to promote autophagy.
Cell death & disease
Wang W,Wang L,Yang M,Wu C,Lan R,Wang W,Li Y
Wed Nov 10 00:00:00 UTC 2021
Autophagy Blockade by Ai Du Qing Formula Promotes Chemosensitivity of Breast Cancer Stem Cells Via GRP78/β-Catenin/ABCG2 Axis.
Frontiers in pharmacology
Liao M,Wang C,Yang B,Huang D,Zheng Y,Wang S,Wang X,Zhang J,Tang C,Xu Z,He Y,Huang R,Zhang F,Wang Z,Wang N
Wed Mar 09 00:00:00 UTC 2022
Maternal selenium deficiency suppresses proliferation, induces autophagy dysfunction and apoptosis in the placenta of mice.
Metallomics : integrated biometal science
Li M,Cheng W,Zhang L
Fri Nov 19 00:00:00 UTC 2021
Pharmacological activation of REV-ERBα improves nonalcoholic steatohepatitis by regulating intestinal permeability.
Metabolism: clinical and experimental
Ni Y,Zhao Y,Ma L,Wang Z,Ni L,Hu L,Fu Z
Fri Jan 01 00:00:00 UTC 2021
Oridonin Alleviates LPS-Induced Depression by Inhibiting NLRP3 Inflammasome via Activation of Autophagy.
Frontiers in medicine
Li C,Zhu Y,Wu Y,Fu M,Wu Y,Wu Y,Qiu Y,Zhang H,Ding M
Tue Feb 01 00:00:00 UTC 2022
Apigenin Alleviates Liver Fibrosis by Inhibiting Hepatic Stellate Cell Activation and Autophagy via TGF-β1/Smad3 and p38/PPARα Pathways.
PPAR research
Ji J,Yu Q,Dai W,Wu L,Feng J,Zheng Y,Li Y,Guo C
Wed Apr 20 00:00:00 UTC 2022
Apigenin Alleviates Liver Fibrosis by Inhibiting Hepatic Stellate Cell Activation and Autophagy via TGF-β1/Smad3 and p38/PPARα Pathways.
PPAR research
Ji J,Yu Q,Dai W,Wu L,Feng J,Zheng Y,Li Y,Guo C
Wed Apr 20 00:00:00 UTC 2022
Apigenin Alleviates Liver Fibrosis by Inhibiting Hepatic Stellate Cell Activation and Autophagy via TGF-β1/Smad3 and p38/PPARα Pathways.
PPAR research
Ji J,Yu Q,Dai W,Wu L,Feng J,Zheng Y,Li Y,Guo C
Wed Apr 20 00:00:00 UTC 2022
Apigenin Alleviates Liver Fibrosis by Inhibiting Hepatic Stellate Cell Activation and Autophagy via TGF-β1/Smad3 and p38/PPARα Pathways.
PPAR research
Ji J,Yu Q,Dai W,Wu L,Feng J,Zheng Y,Li Y,Guo C
Wed Apr 20 00:00:00 UTC 2022
Honokiol induces paraptosis-like cell death of acute promyelocytic leukemia via mTOR & MAPK signaling pathways activation.
Apoptosis : an international journal on programmed cell death
Liu X,Gu Y,Bian Y,Cai D,Li Y,Zhao Y,Zhang Z,Xue M,Zhang L
Thu Apr 01 00:00:00 UTC 2021
Sitagliptin attenuates endothelial dysfunction independent of its blood glucose controlling effect.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology
Chang XM,Xiao F,Pan Q,Wang XX,Guo LX
Wed Sep 01 00:00:00 UTC 2021
Sitagliptin attenuates endothelial dysfunction independent of its blood glucose controlling effect.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology
Chang XM,Xiao F,Pan Q,Wang XX,Guo LX
Wed Sep 01 00:00:00 UTC 2021
Reactive Oxygen Species Mediate 6c-Induced Mitochondrial and Lysosomal Dysfunction, Autophagic Cell Death, and DNA Damage in Hepatocellular Carcinoma.
International journal of molecular sciences
Wang S,Xu X,Che D,Fan R,Gao M,Cao Y,Ge C,Feng Y,Li J,Xie S,Wang C,Dai F,Gao L,Wang Y
Tue Oct 12 00:00:00 UTC 2021
Transcription Factor E2F1 Knockout Promotes Mice White Adipose Tissue Browning Through Autophagy Inhibition.
Frontiers in physiology
Xiong M,Hu W,Tan Y,Yu H,Zhang Q,Zhao C,Yi Y,Wang Y,Wu Y,Wu M
Fri Nov 26 00:00:00 UTC 2021
Effect of GLUT1 Inhibition and Autophagy Modulation on the Growth and Migration of Laryngeal Carcinoma Stem Cells Under Hypoxic and Low-Glucose Conditions.
OncoTargets and therapy
Chen XH,Liu J,Zhong JT,Zhou SH,Fan J
Sat Apr 23 00:00:00 UTC 2022
Genome-scale CRISPRa screening identifies MTX1 as a contributor for sorafenib resistance in hepatocellular carcinoma by augmenting autophagy.
International journal of biological sciences
Li L,Yu S,Hu Q,Hai Y,Li Y
Fri Mar 18 00:00:00 UTC 2022
Biofunctional magnesium-coated Ti6Al4V scaffolds promote autophagy-dependent apoptosis in osteosarcoma by activating the AMPK/mTOR/ULK1 signaling pathway.
Materials today. Bio
Wei X,Tang Z,Wu H,Zuo X,Dong H,Tan L,Wang W,Liu Y,Wu Z,Shi L,Wang N,Li X,Xiao X,Guo Z
Wed Sep 01 00:00:00 UTC 2021
UPF1 impacts on mTOR signaling pathway and autophagy in endometrioid endometrial carcinoma.
Aging
Zhang M,Chen H,Qin P,Cai T,Li L,Chen R,Liu S,Chen H,Lin W,Chen H,Strickland AL,Xiong H,Jiang Q
Tue Sep 14 00:00:00 UTC 2021
Ginsenoside Rb1 alleviates liver injury induced by 3-chloro-1,2-propanediol by stimulating autophagic flux.
Journal of food science
Lu J,Lu J,Bu X,Li Y,Ge G,Guan S
Wed Dec 01 00:00:00 UTC 2021
The SGLT2 inhibitor empagliflozin negatively regulates IL-17/IL-23 axis-mediated inflammatory responses in T2DM with NAFLD via the AMPK/mTOR/autophagy pathway.
International immunopharmacology
Meng Z,Liu X,Li T,Fang T,Cheng Y,Han L,Sun B,Chen L
Sat May 01 00:00:00 UTC 2021
Mitochondrial dysfunction and mitophagy pathway activation in hepatitis E virus-infected livers of Mongolian gerbils.
Virus research
Yang Y,Tian J,Zhang H,Ma M,Li H,Liu T,Yang Y,Liu T,She R
Wed Sep 01 00:00:00 UTC 2021
Mitochondrial dysfunction and mitophagy pathway activation in hepatitis E virus-infected livers of Mongolian gerbils.
Virus research
Yang Y,Tian J,Zhang H,Ma M,Li H,Liu T,Yang Y,Liu T,She R
Wed Sep 01 00:00:00 UTC 2021
Design, Synthesis, and Biological Evaluation of Pyrano[2,3-c]-pyrazole-Based RalA Inhibitors Against Hepatocellular Carcinoma.
Frontiers in chemistry
Wang Y,He M,Li X,Chai J,Jiang Q,Peng C,He G,Huang W
Tue Dec 07 00:00:00 UTC 2021
Design, Synthesis, and Biological Evaluation of Pyrano[2,3-c]-pyrazole-Based RalA Inhibitors Against Hepatocellular Carcinoma.
Frontiers in chemistry
Wang Y,He M,Li X,Chai J,Jiang Q,Peng C,He G,Huang W
Tue Dec 07 00:00:00 UTC 2021
lncRNA NEAT1 ameliorates LPS‑induced inflammation in MG63 cells by activating autophagy and suppressing the NLRP3 inflammasome.
International journal of molecular medicine
Dai W,Wang M,Wang P,Wen J,Wang J,Cha S,Xiao X,He Y,Shu R,Bai D
Mon Feb 01 00:00:00 UTC 2021
The AMPK-MFN2 axis regulates MAM dynamics and autophagy induced by energy stresses.
Autophagy
Hu Y,Chen H,Zhang L,Lin X,Li X,Zhuang H,Fan H,Meng T,He Z,Huang H,Gong Q,Zhu D,Xu Y,He P,Li L,Feng D
Sat May 01 00:00:00 UTC 2021
The AMPK-MFN2 axis regulates MAM dynamics and autophagy induced by energy stresses.
Autophagy
Hu Y,Chen H,Zhang L,Lin X,Li X,Zhuang H,Fan H,Meng T,He Z,Huang H,Gong Q,Zhu D,Xu Y,He P,Li L,Feng D
Sat May 01 00:00:00 UTC 2021
The AMPK-MFN2 axis regulates MAM dynamics and autophagy induced by energy stresses.
Autophagy
Hu Y,Chen H,Zhang L,Lin X,Li X,Zhuang H,Fan H,Meng T,He Z,Huang H,Gong Q,Zhu D,Xu Y,He P,Li L,Feng D
Sat May 01 00:00:00 UTC 2021
The AMPK-MFN2 axis regulates MAM dynamics and autophagy induced by energy stresses.
Autophagy
Hu Y,Chen H,Zhang L,Lin X,Li X,Zhuang H,Fan H,Meng T,He Z,Huang H,Gong Q,Zhu D,Xu Y,He P,Li L,Feng D
Sat May 01 00:00:00 UTC 2021
sTNFRII-Fc modification protects human UC-MSCs against apoptosis/autophagy induced by TNF-α and enhances their efficacy in alleviating inflammatory arthritis.
Stem cell research & therapy
Zhao Y,Yang X,Li S,Zhang B,Li S,Wang X,Wang Y,Jia C,Chang Y,Wei W
Sat Oct 09 00:00:00 UTC 2021
Trimetazidine and exercise provide comparable improvements to high fat diet-induced muscle dysfunction through enhancement of mitochondrial quality control.
Scientific reports
Zhang W,You B,Qi D,Qiu L,Ripley-Gonzalez JW,Zheng F,Fu S,Li C,Dun Y,Liu S
Mon Sep 27 00:00:00 UTC 2021
Myricetin Induces Autophagy and Cell Cycle Arrest of HCC by Inhibiting MARCH1-Regulated Stat3 and p38 MAPK Signaling Pathways.
Frontiers in pharmacology
Yang W,Su J,Li M,Li T,Wang X,Zhao M,Hu X
Fri Nov 05 00:00:00 UTC 2021
Myricetin Induces Autophagy and Cell Cycle Arrest of HCC by Inhibiting MARCH1-Regulated Stat3 and p38 MAPK Signaling Pathways.
Frontiers in pharmacology
Yang W,Su J,Li M,Li T,Wang X,Zhao M,Hu X
Fri Nov 05 00:00:00 UTC 2021
The prohibitin-binding compound fluorizoline inhibits mitophagy in cancer cells.
Oncogenesis
Núñez-Vázquez S,Saura-Esteller J,Sánchez-Vera I,Guilbaud E,Cosialls AM,Pons G,Ricci JE,Iglesias-Serret D,Marchetti S,Gil J
Mon Sep 27 00:00:00 UTC 2021
Inefficient quality control of ribosome stalling during APP synthesis generates CAT-tailed species that precipitate hallmarks of Alzheimer's disease.
Acta neuropathologica communications
Rimal S,Li Y,Vartak R,Geng J,Tantray I,Li S,Huh S,Vogel H,Glabe C,Grinberg LT,Spina S,Seeley WW,Guo S,Lu B
Mon Oct 18 00:00:00 UTC 2021
Babao Dan Reverses Multiple-Drug Resistance in Gastric Cancer Cells via Triggering Apoptosis and Autophagy and Inhibiting PI3K/AKT/mTOR Signaling.
Evidence-based complementary and alternative medicine : eCAM
Zhao J,Lan W,Peng J,Guan B,Liu J,Zhang M,Zhan Z,Lin J
Tue Jul 27 00:00:00 UTC 2021
PPT1 Reduction Contributes to Erianin-Induced Growth Inhibition in Oral Squamous Carcinoma Cells.
Frontiers in cell and developmental biology
Luo Q,Li X,Gan G,Yang M,Chen X,Chen F
Sat Jul 16 00:00:00 UTC 2022
Inhibition of endoplasmic reticulum stress-related autophagy attenuates MCLR-induced apoptosis in zebrafish testis and mouse TM4 cells.
Ecotoxicology and environmental safety
Wu Z,Wang F,Hu L,Zhang J,Chen D,Zhao S
Wed Sep 15 00:00:00 UTC 2021
Lead-induced cardiomyocytes apoptosis by inhibiting gap junction intercellular communication via autophagy activation.
Chemico-biological interactions
Li C,Shi L,Peng C,Yu G,Zhang Y,Du Z
Mon Mar 01 00:00:00 UTC 2021
Characterization of Interplay Between Autophagy and Ferroptosis and Their Synergistical Roles on Manipulating Immunological Tumor Microenvironment in Squamous Cell Carcinomas.
Frontiers in immunology
Chen L,Niu X,Qiao X,Liu S,Ma H,Shi X,He X,Zhong M
Fri Apr 08 00:00:00 UTC 2022
Acupuncture regulates the autophagy of ovarian granulosa cells in polycystic ovarian syndrome ovulation disorder by inhibiting the PI3K/AKT/mTOR pathway through LncMEG3.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Chen X,Tang H,Liang Y,Wu P,Xie L,Ding Y,Yang P,Long B,Lin J
Wed Dec 01 00:00:00 UTC 2021
Acupuncture regulates the autophagy of ovarian granulosa cells in polycystic ovarian syndrome ovulation disorder by inhibiting the PI3K/AKT/mTOR pathway through LncMEG3.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Chen X,Tang H,Liang Y,Wu P,Xie L,Ding Y,Yang P,Long B,Lin J
Wed Dec 01 00:00:00 UTC 2021
Upregulation of thioredoxin contributes to inhibiting diabetic hearing impairment.
Diabetes research and clinical practice
Ren X,Lv J,Fu Y,Zhang N,Zhang C,Dong Z,Chudhary M,Zhong S,Kong L,Kong H
Wed Sep 01 00:00:00 UTC 2021
Atmospheric PM2.5 blocking up autophagic flux in HUVECs via inhibiting Sntaxin-17 and LAMP2.
Ecotoxicology and environmental safety
Liu Y,Zhao D,Peng W,Xue P,Jiang X,Chen S,Gao H,Wang X,Feng S
Fri Jan 15 00:00:00 UTC 2021
Shengma Biejia Decoction Inhibits Cell Growth in Multiple Myeloma by Inducing Autophagy-Mediated Apoptosis Through the ERK/mTOR Pathway.
Frontiers in pharmacology
Dai H,Ma B,Dai X,Pang J,Wang J,Zhao Y,Wang M,Zhang H,Gao H,Qian S,Tian F,Sun X
Fri Apr 16 00:00:00 UTC 2021
Shengma Biejia Decoction Inhibits Cell Growth in Multiple Myeloma by Inducing Autophagy-Mediated Apoptosis Through the ERK/mTOR Pathway.
Frontiers in pharmacology
Dai H,Ma B,Dai X,Pang J,Wang J,Zhao Y,Wang M,Zhang H,Gao H,Qian S,Tian F,Sun X
Fri Apr 16 00:00:00 UTC 2021
Expression of long-chain noncoding RNA GAS5 in osteoarthritis and its effect on apoptosis and autophagy of osteoarthritis chondrocytes.
Histology and histopathology
Ji Q,Qiao X,Liu Y,Wang D
Thu Apr 01 00:00:00 UTC 2021
AKR1B10 negatively regulates autophagy through reducing GAPDH upon glucose starvation in colon cancer.
Journal of cell science
Li W,Liu C,Huang Z,Shi L,Zhong C,Zhou W,Meng P,Li Z,Wang S,Luo F,Yan J,Wu T
Thu Apr 15 00:00:00 UTC 2021
Roles of endoplasmic reticulum stress in 2,2',4,4'-tetrabromodiphenylether-induced thyroid cell apoptosis and autophagy.
Ecotoxicology and environmental safety
Ma R,Li B,Zhang C,Lei R,Zhang X,Wang C,Zhang S,Wang A
Sat May 01 00:00:00 UTC 2021
Cycloastragenol alleviates airway inflammation in asthmatic mice by inhibiting autophagy.
Molecular medicine reports
Zhu X,Cao Y,Su M,Chen M,Li C,Yi L,Qin J,Tulake W,Teng F,Zhong Y,Tang W,Wang S,Dong J
Mon Nov 01 00:00:00 UTC 2021
Cycloastragenol alleviates airway inflammation in asthmatic mice by inhibiting autophagy.
Molecular medicine reports
Zhu X,Cao Y,Su M,Chen M,Li C,Yi L,Qin J,Tulake W,Teng F,Zhong Y,Tang W,Wang S,Dong J
Mon Nov 01 00:00:00 UTC 2021
Delivery of siRNA Using Functionalized Gold Nanorods Enhances Anti-Osteosarcoma Efficacy.
Frontiers in pharmacology
Zhang M,Lin J,Jin J,Yu W,Qi Y,Tao H
Fri Jan 07 00:00:00 UTC 2022
Protective effect of scorpion venom heat-resistant synthetic peptide against PM2.5-induced microglial polarization via TLR4-mediated autophagy activating PI3K/AKT/NF-κB signaling pathway.
Journal of neuroimmunology
Xu X,Xu H,Ren F,Huang L,Xu J,Li F
Tue Jun 15 00:00:00 UTC 2021
Protective effect of Apelin/APJ system on lipopolysaccharide-related cardiac dysfunction.
General physiology and biophysics
Hu J,Huo S,Dou S,Jiang W,Li Y
Sat May 01 00:00:00 UTC 2021
Hsa_circ_0004058 inhibits apoptosis of SRA01/04 cells by promoting autophagy via miR-186/ATG7 axis.
Experimental eye research
Wang Y,Wu Z,Huang Y,Zhang Y
Fri Oct 01 00:00:00 UTC 2021
Cholesterol-associated lysosomal disorder triggers cell death of hematological malignancy: Dynamic analysis on cytotoxic effects of LW-218.
Acta pharmaceutica Sinica. B
Hu P,Li H,Sun W,Wang H,Yu X,Qing Y,Wang Z,Zhu M,Xu J,Guo Q,Hui H
Fri Oct 01 00:00:00 UTC 2021
Cholesterol-associated lysosomal disorder triggers cell death of hematological malignancy: Dynamic analysis on cytotoxic effects of LW-218.
Acta pharmaceutica Sinica. B
Hu P,Li H,Sun W,Wang H,Yu X,Qing Y,Wang Z,Zhu M,Xu J,Guo Q,Hui H
Fri Oct 01 00:00:00 UTC 2021
Neuroprotective Effects of Oxymatrine on PI3K/Akt/mTOR Pathway After Hypoxic-Ischemic Brain Damage in Neonatal Rats.
Frontiers in pharmacology
Wei W,Lu M,Lan XB,Liu N,Su WK,Dushkin AV,Yu JQ
Sun May 02 00:00:00 UTC 2021
Tripterygium glycoside suppresses epithelial‑to‑mesenchymal transition of diabetic kidney disease podocytes by targeting autophagy through the mTOR/Twist1 pathway.
Molecular medicine reports
Tao M,Zheng D,Liang X,Wu D,Hu K,Jin J,He Q
Sun Aug 01 00:00:00 UTC 2021
Tripterygium glycoside suppresses epithelial‑to‑mesenchymal transition of diabetic kidney disease podocytes by targeting autophagy through the mTOR/Twist1 pathway.
Molecular medicine reports
Tao M,Zheng D,Liang X,Wu D,Hu K,Jin J,He Q
Sun Aug 01 00:00:00 UTC 2021
Zirconia Nanoparticles Induce HeLa Cell Death Through Mitochondrial Apoptosis and Autophagy Pathways Mediated by ROS.
Frontiers in chemistry
Shang Y,Wang Q,Li J,Liu H,Zhao Q,Huang X,Dong H,Chen W,Gui R,Nie X
Sat Apr 03 00:00:00 UTC 2021
Vildagliptin Attenuates Myocardial Dysfunction and Restores Autophagy via miR-21/SPRY1/ERK in Diabetic Mice Heart.
Frontiers in pharmacology
Li X,Meng C,Han F,Yang J,Wang J,Zhu Y,Cui X,Zuo M,Xu J,Chang B
Tue Apr 06 00:00:00 UTC 2021
1,3-dichloro-2-propanol induced lipid accumulation by blocking autophagy flux in HepG2 cells.
Toxicology
Lu J,Fan Y,Liu M,Zhang Q,Guan S
Fri Apr 30 00:00:00 UTC 2021
1,3-dichloro-2-propanol induced lipid accumulation by blocking autophagy flux in HepG2 cells.
Toxicology
Lu J,Fan Y,Liu M,Zhang Q,Guan S
Fri Apr 30 00:00:00 UTC 2021
Oroxylin A is a severe acute respiratory syndrome coronavirus 2-spiked pseudotyped virus blocker obtained from Radix Scutellariae using angiotensin-converting enzyme II/cell membrane chromatography.
Phytotherapy research : PTR
Gao J,Ding Y,Wang Y,Liang P,Zhang L,Liu R
Tue Jun 01 00:00:00 UTC 2021
Transcriptome-Based Network Analysis Reveals Hirudin Potentiates Anti-Renal Fibrosis Efficacy in UUO Rats.
Frontiers in pharmacology
Yu HX,Lin W,Yang K,Wei LJ,Chen JL,Liu XY,Zhong K,Chen X,Pei M,Yang HT
Sat Oct 09 00:00:00 UTC 2021
OAB-14 Effectively Ameliorates the Dysfunction of the Endosomal-Autophagic-Lysosomal Pathway in APP/PS1 Transgenic Mice.
ACS chemical neuroscience
Guo X,Bao X,Wang X,Liu D,Liu P,Chi T,Ji X,Zheng Z,Chen G,Zou L
Wed Nov 03 00:00:00 UTC 2021
Hepatic COX-2 expression protects mice from an alcohol-high fat diet-induced metabolic disorder by involving protein acetylation related energy metabolism.
Alcohol (Fayetteville, N.Y.)
Chen M,Sun X,Wei W,Cucarella C,Martín-Sanz P,Casado M,Pi L,Ren B,Cao Q
Sat May 01 00:00:00 UTC 2021
Stimulation of AMPK Prevents Diabetes-Induced Photoreceptor Cell Degeneration.
Oxidative medicine and cellular longevity
Song S,Bao S,Zhang C,Zhang J,Lv J,Li X,Chudhary M,Ren X,Kong L
Tue Dec 28 00:00:00 UTC 2021
Silencing of circIgf1r plays a protective role in neuronal injury via regulating astrocyte polarization during epilepsy.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Shao L,Jiang GT,Yang XL,Zeng ML,Cheng JJ,Kong S,Dong X,Chen TX,Han S,Yin J,Liu WH,He XH,He C,Peng BW
Mon Feb 01 00:00:00 UTC 2021
Silencing of circIgf1r plays a protective role in neuronal injury via regulating astrocyte polarization during epilepsy.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Shao L,Jiang GT,Yang XL,Zeng ML,Cheng JJ,Kong S,Dong X,Chen TX,Han S,Yin J,Liu WH,He XH,He C,Peng BW
Mon Feb 01 00:00:00 UTC 2021
Bone marrow-derived mesenchymal stem cells attenuate silica-induced pulmonary fibrosis by inhibiting apoptosis and pyroptosis but not autophagy in rats.
Ecotoxicology and environmental safety
Zhao Q,Hao C,Wei J,Huang R,Li C,Yao W
Sat Apr 10 00:00:00 UTC 2021
Epimedokoreanin C, a prenylated flavonoid isolated from Epimedium koreanum, induces non-apoptotic cell death with the characteristics of methuosis in lung cancer cells.
American journal of cancer research
Liu X,Wang S,Zheng H,Liu Q,Shen T,Wang X,Ren D
Sat Aug 07 00:00:00 UTC 2021
The Disulfiram/Copper Complex Induces Autophagic Cell Death in Colorectal Cancer by Targeting ULK1.
Frontiers in pharmacology
Hu Y,Qian Y,Wei J,Jin T,Kong X,Cao H,Ding K
Sat Dec 11 00:00:00 UTC 2021
The Disulfiram/Copper Complex Induces Autophagic Cell Death in Colorectal Cancer by Targeting ULK1.
Frontiers in pharmacology
Hu Y,Qian Y,Wei J,Jin T,Kong X,Cao H,Ding K
Sat Dec 11 00:00:00 UTC 2021
The Disulfiram/Copper Complex Induces Autophagic Cell Death in Colorectal Cancer by Targeting ULK1.
Frontiers in pharmacology
Hu Y,Qian Y,Wei J,Jin T,Kong X,Cao H,Ding K
Sat Dec 11 00:00:00 UTC 2021
The Disulfiram/Copper Complex Induces Autophagic Cell Death in Colorectal Cancer by Targeting ULK1.
Frontiers in pharmacology
Hu Y,Qian Y,Wei J,Jin T,Kong X,Cao H,Ding K
Sat Dec 11 00:00:00 UTC 2021
The Difference in Expression of Autophagy-Related Proteins in Lesional and Perilesional Skin in Adult Patients with Active and Stable Generalized Vitiligo-A Cross-Sectional Pilot Study.
Indian journal of dermatology
Yu H,Lin X,Huang Y,Cheng H,Seifert O
Thu Apr 28 00:00:00 UTC 2022
Compensatory combination of mTOR and TrxR inhibitors to cause oxidative stress and regression of tumors.
Theranostics
Xia Y,Chen J,Yu Y,Wu F,Shen X,Qiu C,Zhang T,Hong L,Zheng P,Shao R,Xu C,Wu F,Chen W,Xie C,Cui R,Zou P
Thu Jul 29 00:00:00 UTC 2021
Alleviation of CCCP-induced mitochondrial injury by augmenter of liver regeneration via the PINK1/Parkin pathway-dependent mitophagy.
Experimental cell research
Zhang J,Chen S,Li Y,Xiao W,An W
Wed Dec 01 00:00:00 UTC 2021
W436, a novel SMART derivative, exhibits anti-hepatocarcinoma activity by inducing apoptosis and G2/M cell cycle arrest in vitro and in vivo and induces protective autophagy.
Journal of biochemical and molecular toxicology
Man S,Wu Z,Sun R,Guan Q,Li Z,Zuo D,Zhang W,Wu Y
Sun Aug 01 00:00:00 UTC 2021
25-Hydroxycholesterol mitigates hepatic ischemia reperfusion injury via mediating mitophagy.
International immunopharmacology
Cao Q,Luo J,Xiong Y,Liu Z,Ye Q
Thu Jul 01 00:00:00 UTC 2021
Pterostilbene alleviates pulmonary fibrosis by regulating ASIC2.
Chinese medicine
Peng Y,Zhang Y,Zhang Y,Wang X,Xia Y
Wed Jul 28 00:00:00 UTC 2021
Inhibition of autophagy with Chloroquine enhanced apoptosis induced by 5-aminolevulinic acid-photodynamic therapy in secondary hyperparathyroidism primary cells and organoids.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zeng L,Zou Q,Huang P,Xiong L,Cheng Y,Chen Q,Li Y,He H,Yi W,Wei W
Fri Oct 01 00:00:00 UTC 2021
Inhibition of autophagy with Chloroquine enhanced apoptosis induced by 5-aminolevulinic acid-photodynamic therapy in secondary hyperparathyroidism primary cells and organoids.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zeng L,Zou Q,Huang P,Xiong L,Cheng Y,Chen Q,Li Y,He H,Yi W,Wei W
Fri Oct 01 00:00:00 UTC 2021
Melatonin induces the rejuvenation of long-term ex vivo expanded periodontal ligament stem cells by modulating the autophagic process.
Stem cell research & therapy
Tan YZ,Xu XY,Dai JM,Yin Y,He XT,Zhang YL,Zhu TX,An Y,Tian BM,Chen FM
Thu Apr 29 00:00:00 UTC 2021
Catalpol-Induced AMPK Activation Alleviates Cisplatin-Induced Nephrotoxicity through the Mitochondrial-Dependent Pathway without Compromising Its Anticancer Properties.
Oxidative medicine and cellular longevity
Zhang J,Zhao T,Wang C,Meng Q,Huo X,Wang C,Sun P,Sun H,Ma X,Wu J,Liu K
Tue Sep 07 00:00:00 UTC 2021
ERRFI1 induces apoptosis of hepatocellular carcinoma cells in response to tryptophan deficiency.
Cell death discovery
Cui M,Liu D,Xiong W,Wang Y,Mi J
Mon Oct 04 00:00:00 UTC 2021
Autocrined leptin promotes proliferation of non-small cell lung cancer (NSCLC) via PI3K/AKT and p53 pathways.
Annals of translational medicine
Wang J,Zhou F,Li F,Wang B,Hu Y,Li X
Thu Apr 01 00:00:00 UTC 2021
Relieving lipid accumulation through UCP1 suppresses the progression of acute kidney injury by promoting the AMPK/ULK1/autophagy pathway.
Theranostics
Xiong W,Xiong Z,Song A,Lei C,Ye C,Zhang C
Mon Aug 16 00:00:00 UTC 2021
Uhrf1 regulates H3K9me2 modification of mTOR to inhibit the effect of autophagy in myocardial ischemia-reperfusion injury.
Aging
Li HG,Tian WH,Qin CL,Ye RR,Liu DH,Liu HW
Fri Mar 19 00:00:00 UTC 2021
Circular RNA TRAPPC6B inhibits intracellular Mycobacterium tuberculosis growth while inducing autophagy in macrophages by targeting microRNA-874-3p.
Clinical & translational immunology
Luo HL,Pi J,Zhang JA,Yang EZ,Xu H,Luo H,Shen L,Peng Y,Liu GB,Song CM,Li KY,Wu XJ,Zheng BY,Shen HB,Chen ZW,Xu JF
Thu Apr 21 00:00:00 UTC 2022
Mild hypothermia pretreatment extenuates liver ischemia-reperfusion injury through Rab7-mediated autophagosomes-lysosomes fusion.
Biochemical and biophysical research communications
Liu A,Wang W,Lu Z,Liu Z,Zhou W,Zhong Z,Ye Q
Fri Apr 23 00:00:00 UTC 2021
Mild hypothermia pretreatment extenuates liver ischemia-reperfusion injury through Rab7-mediated autophagosomes-lysosomes fusion.
Biochemical and biophysical research communications
Liu A,Wang W,Lu Z,Liu Z,Zhou W,Zhong Z,Ye Q
Fri Apr 23 00:00:00 UTC 2021
Downregulation of miR-29c promotes muscle wasting by modulating the activity of leukemia inhibitory factor in lung cancer cachexia.
Cancer cell international
Xie K,Xiong H,Xiao W,Xiong Z,Hu W,Ye J,Xu N,Shi J,Yuan C,Chen Z,Miao D,Zhang X,Yang H
Sat Nov 27 00:00:00 UTC 2021
Claudin-5 Affects Endothelial Autophagy in Response to Early Hypoxia.
Frontiers in physiology
Yu P,Li Y,Zhong G,Li W,Chen B,Zhang J
Sat Sep 18 00:00:00 UTC 2021
FV-429 induces autophagy blockage and lysosome-dependent cell death of T-cell malignancies via lysosomal dysregulation.
Cell death & disease
Hu P,Wang J,Qing Y,Li H,Sun W,Yu X,Hui H,Guo Q,Xu J
Wed Jan 13 00:00:00 UTC 2021
FV-429 induces autophagy blockage and lysosome-dependent cell death of T-cell malignancies via lysosomal dysregulation.
Cell death & disease
Hu P,Wang J,Qing Y,Li H,Sun W,Yu X,Hui H,Guo Q,Xu J
Wed Jan 13 00:00:00 UTC 2021
Cholecalciferol (vitamin D3) has a direct protective activity against interleukin 6-induced atrophy in C2C12 myotubes.
Aging
Teixeira MA,De Feudis M,Reano S,Raiteri T,Scircoli A,Zaggia I,Ruga S,Salvadori L,Prodam F,Marzullo P,Molinari C,Corà D,Filigheddu N
Mon Feb 22 00:00:00 UTC 2021
The Dual Histone Deacetylase-Proteasome Inhibitor RTS-V5 Acts Synergistically With Ritonavir to Induce Endoplasmic Reticulum Stress in Bladder Cancer Cells.
Anticancer research
Okubo K,REßING N,Schulz WA,Hansen FK,Asano T,Sato A
Wed Dec 01 00:00:00 UTC 2021
Esomeprazole overcomes paclitaxel-resistance and enhances anticancer effects of paclitaxel by inducing autophagy in A549/Taxol cells.
Cell biology international
Bai Z,Ding N,Ge J,Wang Y,Wang L,Wu N,Wei Q,Xu S,Liu X,Zhou G
Fri Jan 01 00:00:00 UTC 2021
Curcumin is a Potential Adjuvant to Alleviates Diabetic Retinal Injury via Reducing Oxidative Stress and Maintaining Nrf2 Pathway Homeostasis.
Frontiers in pharmacology
Xie T,Chen X,Chen W,Huang S,Peng X,Tian L,Wu X,Huang Y
Tue Dec 28 00:00:00 UTC 2021
Hydrogen sulfide ameliorates doxorubicin‑induced myocardial fibrosis in rats via the PI3K/AKT/mTOR pathway.
Molecular medicine reports
Nie L,Liu M,Chen J,Wu Q,Li Y,Yi J,Zheng X,Zhang J,Chu C,Yang J
Thu Apr 01 00:00:00 UTC 2021
Screening and identification of NOTCH1, CDKN2A, and NOS3 as differentially expressed autophagy-related genes in erectile dysfunction.
PeerJ
Luo C,Zhou X,Wang L,Zeng Q,Fan J,He S,Zhang H,Wei A
Tue Apr 26 00:00:00 UTC 2022
Screening and identification of NOTCH1, CDKN2A, and NOS3 as differentially expressed autophagy-related genes in erectile dysfunction.
PeerJ
Luo C,Zhou X,Wang L,Zeng Q,Fan J,He S,Zhang H,Wei A
Tue Apr 26 00:00:00 UTC 2022
Oxalate Activates Autophagy to Induce Ferroptosis of Renal Tubular Epithelial Cells and Participates in the Formation of Kidney Stones.
Oxidative medicine and cellular longevity
Song Q,Liao W,Chen X,He Z,Li D,Li B,Liu J,Liu L,Xiong Y,Song C,Yang S
Wed Feb 02 00:00:00 UTC 2022
Vitamin D decreases Porphyromonas gingivalis internalized into macrophages by promoting autophagy.
Oral diseases
Niu L,Chen S,Yang X,Ma C,Pan C,Wang H,Li Q,Geng F,Tang X
Fri Oct 01 00:00:00 UTC 2021
Vitamin D decreases Porphyromonas gingivalis internalized into macrophages by promoting autophagy.
Oral diseases
Niu L,Chen S,Yang X,Ma C,Pan C,Wang H,Li Q,Geng F,Tang X
Fri Oct 01 00:00:00 UTC 2021
PPARγ Plays an Important Role in Acute Hepatic Ischemia-Reperfusion Injury via AMPK/mTOR Pathway.
PPAR research
Wu L,Yu Q,Cheng P,Guo C
Mon Apr 25 00:00:00 UTC 2022
Liquiritigenin protects against arsenic trioxide-induced liver injury by inhibiting oxidative stress and enhancing mTOR-mediated autophagy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang M,Xue Y,Zheng B,Li L,Chu X,Zhao Y,Wu Y,Zhang J,Han X,Wu Z,Chu L
Mon Nov 01 00:00:00 UTC 2021
Luteolin attenuates angiotensin II‑induced renal damage in apolipoprotein E‑deficient mice.
Molecular medicine reports
Liu YS,Yang Q,Li S,Luo L,Liu HY,Li XY,Gao ZN
Mon Feb 01 00:00:00 UTC 2021
Luteolin attenuates angiotensin II‑induced renal damage in apolipoprotein E‑deficient mice.
Molecular medicine reports
Liu YS,Yang Q,Li S,Luo L,Liu HY,Li XY,Gao ZN
Mon Feb 01 00:00:00 UTC 2021
Discovery of First-in-Class Dual PARP and EZH2 Inhibitors for Triple-Negative Breast Cancer with Wild-Type BRCA.
Journal of medicinal chemistry
Wang C,Qu L,Li S,Yin F,Ji L,Peng W,Luo H,Lu D,Liu X,Chen X,Kong L,Wang X
Thu Sep 09 00:00:00 UTC 2021
Discovery of First-in-Class Dual PARP and EZH2 Inhibitors for Triple-Negative Breast Cancer with Wild-Type BRCA.
Journal of medicinal chemistry
Wang C,Qu L,Li S,Yin F,Ji L,Peng W,Luo H,Lu D,Liu X,Chen X,Kong L,Wang X
Thu Sep 09 00:00:00 UTC 2021
Combination effect of curcumin with docetaxel on the PI3K/AKT/mTOR pathway to induce autophagy and apoptosis in esophageal squamous cell carcinoma.
American journal of translational research
Deng L,Wu X,Zhu X,Yu Z,Liu Z,Wang J,Zheng Y
Wed Feb 03 00:00:00 UTC 2021
Compound essential oils relieve oxidative stress caused by PM2 .5 exposure by inhibiting autophagy through the AMPK/mTOR pathway.
Environmental toxicology
Ren F,Xu X,Xu J,Mei Y,Zhang J,Wang X,Li F
Wed Sep 01 00:00:00 UTC 2021
Compound essential oils relieve oxidative stress caused by PM2 .5 exposure by inhibiting autophagy through the AMPK/mTOR pathway.
Environmental toxicology
Ren F,Xu X,Xu J,Mei Y,Zhang J,Wang X,Li F
Wed Sep 01 00:00:00 UTC 2021
Ferritinophagy-Mediated ROS Production Contributed to Proliferation Inhibition, Apoptosis, and Ferroptosis Induction in Action of Mechanism of 2-Pyridylhydrazone Dithiocarbamate Acetate.
Oxidative medicine and cellular longevity
Li L,Li H,Li Y,Feng J,Guan D,Zhang Y,Fu Y,Li S,Li C
Thu Feb 17 00:00:00 UTC 2022
Identification of Potentially Related Genes and Mechanisms Involved in Skeletal Muscle Atrophy Induced by Excessive Exercise in Zebrafish.
Biology
Sun CC,Zhou ZQ,Chen ZL,Zhu RK,Yang D,Peng XY,Zheng L,Tang CF
Tue Aug 10 00:00:00 UTC 2021
Sanguinarine impairs lysosomal function and induces ROS-dependent mitophagy and apoptosis in human hepatocellular carcinoma cells.
Archives of pharmacal research
Wang J,Su Q,Wu Q,Chen K,Ullah A,Ghauri MA,Zhang Y
Mon Nov 01 00:00:00 UTC 2021
Sanguinarine impairs lysosomal function and induces ROS-dependent mitophagy and apoptosis in human hepatocellular carcinoma cells.
Archives of pharmacal research
Wang J,Su Q,Wu Q,Chen K,Ullah A,Ghauri MA,Zhang Y
Mon Nov 01 00:00:00 UTC 2021
Sanguinarine impairs lysosomal function and induces ROS-dependent mitophagy and apoptosis in human hepatocellular carcinoma cells.
Archives of pharmacal research
Wang J,Su Q,Wu Q,Chen K,Ullah A,Ghauri MA,Zhang Y
Mon Nov 01 00:00:00 UTC 2021
TCDD induced lipid accumulation by impairment of autophagic flux in THP-1 macrophages.
Environmental science and pollution research international
Lu J,Liu M,Fan Y,Zheng H,Guan S
Thu Jul 01 00:00:00 UTC 2021
TCDD induced lipid accumulation by impairment of autophagic flux in THP-1 macrophages.
Environmental science and pollution research international
Lu J,Liu M,Fan Y,Zheng H,Guan S
Thu Jul 01 00:00:00 UTC 2021
Autophagy-associated circular RNA hsa_circ_0007813 modulates human bladder cancer progression via hsa-miR-361-3p/IGF2R regulation.
Cell death & disease
Zhang Z,Mou Z,Xu C,Wu S,Dai X,Chen X,Ou Y,Chen Y,Yang C,Jiang H
Sat Aug 07 00:00:00 UTC 2021
Autophagy-associated circular RNA hsa_circ_0007813 modulates human bladder cancer progression via hsa-miR-361-3p/IGF2R regulation.
Cell death & disease
Zhang Z,Mou Z,Xu C,Wu S,Dai X,Chen X,Ou Y,Chen Y,Yang C,Jiang H
Sat Aug 07 00:00:00 UTC 2021
Fibroblast growth factor 21 (FGF21) alleviates senescence, apoptosis, and extracellular matrix degradation in osteoarthritis via the SIRT1-mTOR signaling pathway.
Cell death & disease
Lu H,Jia C,Wu D,Jin H,Lin Z,Pan J,Li X,Wang W
Thu Sep 23 00:00:00 UTC 2021
Fibroblast growth factor 21 (FGF21) alleviates senescence, apoptosis, and extracellular matrix degradation in osteoarthritis via the SIRT1-mTOR signaling pathway.
Cell death & disease
Lu H,Jia C,Wu D,Jin H,Lin Z,Pan J,Li X,Wang W
Thu Sep 23 00:00:00 UTC 2021
Fibroblast growth factor 21 (FGF21) alleviates senescence, apoptosis, and extracellular matrix degradation in osteoarthritis via the SIRT1-mTOR signaling pathway.
Cell death & disease
Lu H,Jia C,Wu D,Jin H,Lin Z,Pan J,Li X,Wang W
Thu Sep 23 00:00:00 UTC 2021
Comparison of toxicity of Ti3 C2 and Nb2 C Mxene quantum dots (QDs) to human umbilical vein endothelial cells.
Journal of applied toxicology : JAT
Gu M,Dai Z,Yan X,Ma J,Niu Y,Lan W,Wang X,Xu Q
Sat May 01 00:00:00 UTC 2021
The nicotinamide phosphoribosyltransferase antagonist FK866 inhibits growth of prostate tumour spheroids and increases doxorubicin retention without changes in drug transporter and cancer stem cell protein expression.
Clinical and experimental pharmacology & physiology
Sauer H,Kampmann H,Khosravi F,Sharifpanah F,Wartenberg M
Mon Mar 01 00:00:00 UTC 2021
The nicotinamide phosphoribosyltransferase antagonist FK866 inhibits growth of prostate tumour spheroids and increases doxorubicin retention without changes in drug transporter and cancer stem cell protein expression.
Clinical and experimental pharmacology & physiology
Sauer H,Kampmann H,Khosravi F,Sharifpanah F,Wartenberg M
Mon Mar 01 00:00:00 UTC 2021
Effect of Vitamin D Deficiency and Supplementation in Lactation and Early Life on Allergic Airway Inflammation and the Expression of Autophagy-Related Genes in an Ovalbumin Mouse Model.
Journal of inflammation research
Zhou Y,Xue Y,Bao A,Han L,Bao W,Xia C,Tian X,Zhang M
Tue Apr 26 00:00:00 UTC 2022
Aquaporin-9 is involved in the lipid-lowering activity of the nutraceutical silybin on hepatocytes through modulation of autophagy and lipid droplets composition.
Biochimica et biophysica acta. Molecular and cell biology of lipids
Baldini F,Portincasa P,Grasselli E,Damonte G,Salis A,Bonomo M,Florio M,Serale N,Voci A,Gena P,Vergani L,Calamita G
Sun Mar 01 00:00:00 UTC 2020
Discovery of novel akt1 inhibitor induces autophagy associated death in hepatocellular carcinoma cells.
European journal of medicinal chemistry
Yu M,Zeng M,Pan Z,Wu F,Guo L,He G
Sun Mar 01 00:00:00 UTC 2020
ChREBP-β regulates thermogenesis in brown adipose tissue.
The Journal of endocrinology
Wei C,Ma X,Su K,Qi S,Zhu Y,Lin J,Wang C,Yang R,Chen X,Wang W,Zhang WJ
Mon Jun 01 00:00:00 UTC 2020
3-Chloro-1, 2-propanediol inhibits autophagic flux by impairment of lysosomal function in HepG2 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Lu J,Lu J,Chen Y,Feng Z,Liu S,Guan S
Thu Oct 01 00:00:00 UTC 2020
MCP-1 mediates ischemia-reperfusion-induced cardiomyocyte apoptosis via MCPIP1 and CaSR.
American journal of physiology. Heart and circulatory physiology
Zhang W,Zhu T,Chen L,Luo W,Chao J
Wed Jan 01 00:00:00 UTC 2020
Tubeimoside I Inhibits Cell Proliferation and Induces a Partly Disrupted and Cytoprotective Autophagy Through Rapidly Hyperactivation of MEK1/2-ERK1/2 Cascade via Promoting PTP1B in Melanoma.
Frontiers in cell and developmental biology
Du J,Dong Z,Tan L,Tan M,Zhang F,Zhang K,Pan G,Li C,Shi S,Zhang Y,Liu Y,Cui H
Tue Jan 05 00:00:00 UTC 2021
Exercise enhances mitochondrial fission and mitophagy to improve myopathy following critical limb ischemia in elderly mice via the PGC1a/FNDC5/irisin pathway.
Skeletal muscle
He W,Wang P,Chen Q,Li C
Tue Sep 15 00:00:00 UTC 2020
Resveratrol inhibits hypertrophic scars formation by activating autophagy via the miR-4654/Rheb axis.
Molecular medicine reports
Pang K,Li B,Tang Z,Yang W,Hao L,Shi Z,Zhang J,Cai L,Li R,Liu Y,Lv Q,Ding J,Han C
Thu Oct 01 00:00:00 UTC 2020
Exogenous H2S Promoted USP8 Sulfhydration to Regulate Mitophagy in the Hearts of db/db Mice.
Aging and disease
Sun Y,Lu F,Yu X,Wang B,Chen J,Lu F,Peng S,Sun X,Yu M,Chen H,Wang Y,Zhang L,Liu N,Du H,Zhao D,Zhang W
Wed Apr 01 00:00:00 UTC 2020
Toll-Like Receptor 2-Mediated Autophagy Promotes Microglial Cell Death by Modulating the Microglial M1/M2 Phenotype.
Inflammation
Ma K,Guo J,Wang G,Ni Q,Liu X
Wed Apr 01 00:00:00 UTC 2020
Disulfiram Chelated With Copper Inhibits the Growth of Gastric Cancer Cells by Modulating Stress Response and Wnt/β-catenin Signaling.
Frontiers in oncology
Wang L,Chai X,Wan R,Zhang H,Zhou C,Xiang L,Paul ME,Li Y
Mon Jan 11 00:00:00 UTC 2021
Nbr1-regulated autophagy in Lactoferrin-induced osteoblastic differentiation.
Bioscience, biotechnology, and biochemistry
Zhang Y,Zhang ZN,Li N,Zhao LJ,Xue Y,Wu HJ,Hou JM
Mon Jun 01 00:00:00 UTC 2020
Calcitriol decreases live Porphyromonas gingivalis internalized into epithelial cells and monocytes by promoting autophagy.
Journal of periodontology
Hu X,Niu L,Ma C,Huang Y,Yang X,Shi Y,Pan C,Liu J,Wang H,Li Q,Geng F,Tang X
Wed Jul 01 00:00:00 UTC 2020
SPAG5-AS1 inhibited autophagy and aggravated apoptosis of podocytes via SPAG5/AKT/mTOR pathway.
Cell proliferation
Xu J,Deng Y,Wang Y,Sun X,Chen S,Fu G
Sat Feb 01 00:00:00 UTC 2020
1-(4-((5-chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)-3-methoxyphenyl)-3-(2-(dimethylamino)ethyl)imidazolidin-2-one (ZX-42), a novel ALK inhibitor, induces apoptosis and protective autophagy in H2228 cells.
The Journal of pharmacy and pharmacology
Wang L,Xu X,Liu T,Wang J,Shen J,Guo M,Wu Y,Zhai X,Zuo D
Thu Oct 01 00:00:00 UTC 2020
Organic anion transporters and PI3K-AKT-mTOR pathway mediate the synergistic anticancer effect of pemetrexed and rhein.
Journal of cellular physiology
Bu T,Wang C,Jin H,Meng Q,Huo X,Sun H,Sun P,Wu J,Ma X,Liu Z,Liu K
Wed Apr 01 00:00:00 UTC 2020
Methylmercury disrupts autophagic flux by inhibiting autophagosome-lysosome fusion in mouse germ cells.
Ecotoxicology and environmental safety
Chen N,Tang X,Ye Z,Wang S,Xiao X
Wed Jul 15 00:00:00 UTC 2020
Methylmercury disrupts autophagic flux by inhibiting autophagosome-lysosome fusion in mouse germ cells.
Ecotoxicology and environmental safety
Chen N,Tang X,Ye Z,Wang S,Xiao X
Wed Jul 15 00:00:00 UTC 2020
TAF1L promotes development of oral squamous cell carcinoma via decreasing autophagy-dependent apoptosis.
International journal of biological sciences
Wang D,Qi H,Zhang H,Zhou W,Li Y,Li A,Liu Q,Wang Y
Thu Sep 30 00:00:00 UTC 2021
TAF1L promotes development of oral squamous cell carcinoma via decreasing autophagy-dependent apoptosis.
International journal of biological sciences
Wang D,Qi H,Zhang H,Zhou W,Li Y,Li A,Liu Q,Wang Y
Thu Sep 30 00:00:00 UTC 2021
A spirostanol saponin isolated from Tupistra chinensis Baker simultaneously induces apoptosis and autophagy by regulating the JNK pathway in human gastric cancer cells.
Steroids
Xu J,Wang Z,Huang Y,Wang Y,Xiang L,He X
Tue Dec 01 00:00:00 UTC 2020
Septin4 as an autophagy modulator regulates Angiotensin-II mediated VSMCs proliferation and migration.
Biochemical and biophysical research communications
Wang N,Xu F,Lu S,Zhang N,Sun Y
Thu Apr 30 00:00:00 UTC 2020
Apigenin induced autophagy and stimulated autophagic lipid degradation.
Food & function
Lu J,Meng Z,Chen Y,Yu L,Gao B,Zheng Y,Guan S
Wed Oct 21 00:00:00 UTC 2020
Apigenin induced autophagy and stimulated autophagic lipid degradation.
Food & function
Lu J,Meng Z,Chen Y,Yu L,Gao B,Zheng Y,Guan S
Wed Oct 21 00:00:00 UTC 2020
BDH2 triggers ROS-induced cell death and autophagy by promoting Nrf2 ubiquitination in gastric cancer.
Journal of experimental & clinical cancer research : CR
Liu JZ,Hu YL,Feng Y,Jiang Y,Guo YB,Liu YF,Chen X,Yang JL,Chen YY,Mao QS,Xue WJ
Tue Jun 30 00:00:00 UTC 2020
CKIP-1 augments autophagy in steatotic hepatocytes by inhibiting Akt/mTOR signal pathway.
Experimental cell research
Li L,Xie P,Lin W,Liu J,Chen J,Guo Z,Bin C,An W,Zhang C,Zhan Y
Tue Dec 01 00:00:00 UTC 2020
Desloratadine, a Novel Antigrowth Reagent for Bladder Cancer.
Technology in cancer research & treatment
Ma J,Qi J,Li S,Zhang C,Wang H,Shao L,Yuan X,Sha Q
Mon Jan 04 00:00:00 UTC 2021
Aconitine induces cardiomyocyte damage by mitigating BNIP3-dependent mitophagy and the TNFα-NLRP3 signalling axis.
Cell proliferation
Peng F,Zhang N,Wang C,Wang X,Huang W,Peng C,He G,Han B
Wed Jan 01 00:00:00 UTC 2020
Aconitine induces cardiomyocyte damage by mitigating BNIP3-dependent mitophagy and the TNFα-NLRP3 signalling axis.
Cell proliferation
Peng F,Zhang N,Wang C,Wang X,Huang W,Peng C,He G,Han B
Wed Jan 01 00:00:00 UTC 2020
NOD2/c-Jun NH2-Terminal Kinase Triggers Mycoplasma ovipneumoniae-Induced Macrophage Autophagy.
Journal of bacteriology
Luo H,Wu X,Xu Z,Hao X,Wang Y,Li M
Wed Sep 23 00:00:00 UTC 2020
Qi Dan Li Xin pill improves chronic heart failure by regulating mTOR/p70S6k-mediated autophagy and inhibiting apoptosis.
Scientific reports
Shi B,Huang Y,Ni J,Chen J,Wei J,Gao H,Li L,Zhou Z,Wang Y,Xu Y,Xu Z,Mao J,Fan G
Wed Apr 08 00:00:00 UTC 2020
Bazedoxifene inhibits PDGF-BB induced VSMC phenotypic switch via regulating the autophagy level.
Life sciences
Song W,Gao K,Huang P,Tang Z,Nie F,Jia S,Guo R
Thu Oct 15 00:00:00 UTC 2020
Autophagy Is a Potential Therapeutic Target Against Duck Tembusu Virus Infection in vivo.
Frontiers in cellular and infection microbiology
Hu Z,Pan Y,Cheng A,Zhang X,Wang M,Chen S,Zhu D,Liu M,Yang Q,Wu Y,Zhao X,Huang J,Zhang S,Mao S,Ou X,Yu Y,Zhang L,Liu Y,Tian B,Pan L,Rehman MU,Yin Z,Jia R
Thu Jun 17 00:00:00 UTC 2021
The c-Myc/miR-27b-3p/ATG10 regulatory axis regulates chemoresistance in colorectal cancer.
Theranostics
Sun W,Li J,Zhou L,Han J,Liu R,Zhang H,Ning T,Gao Z,Liu B,Chen X,Ba Y
Wed Apr 14 00:00:00 UTC 2021
Phosphoproteomics identifies dual-site phosphorylation in an extended basophilic motif regulating FILIP1-mediated degradation of filamin-C.
Communications biology
Reimann L,Schwäble AN,Fricke AL,Mühlhäuser WWD,Leber Y,Lohanadan K,Puchinger MG,Schäuble S,Faessler E,Wiese H,Reichenbach C,Knapp B,Peikert CD,Drepper F,Hahn U,Kreutz C,van der Ven PFM,Radziwill G,Djinović-Carugo K,Fürst DO,Warscheid B
Fri May 22 00:00:00 UTC 2020
Pretreatment with the ALDH2 activator Alda‑1 protects rat livers from ischemia/reperfusion injury by inducing autophagy.
Molecular medicine reports
Liu Z,Ye S,Zhong X,Wang W,Lai CH,Yang W,Yue P,Luo J,Huang X,Zhong Z,Xiong Y,Fan X,Li L,Wang Y,Ye Q
Tue Sep 01 00:00:00 UTC 2020
PDK4 promotes vascular calcification by interfering with autophagic activity and metabolic reprogramming.
Cell death & disease
Ma WQ,Sun XJ,Zhu Y,Liu NF
Tue Nov 17 00:00:00 UTC 2020
CACNA1H downregulation induces skeletal muscle atrophy involving endoplasmic reticulum stress activation and autophagy flux blockade.
Cell death & disease
Li S,Hao M,Li B,Chen M,Chen J,Tang J,Hong S,Min J,Hu M,Hong L
Fri Apr 24 00:00:00 UTC 2020
CACNA1H downregulation induces skeletal muscle atrophy involving endoplasmic reticulum stress activation and autophagy flux blockade.
Cell death & disease
Li S,Hao M,Li B,Chen M,Chen J,Tang J,Hong S,Min J,Hu M,Hong L
Fri Apr 24 00:00:00 UTC 2020
Sanguisorba officinalis L. Suppresses Triple-Negative Breast Cancer Metastasis by Inhibiting Late-Phase Autophagy via Hif-1α/Caveolin-1 Signaling.
Frontiers in pharmacology
Wang N,Muhetaer G,Zhang X,Yang B,Wang C,Zhang Y,Wang X,Zhang J,Wang S,Zheng Y,Zhang F,Wang Z
Mon Jan 24 00:00:00 UTC 2022
Cells deficient for Krüppel-like factor 4 exhibit mitochondrial dysfunction and impaired mitophagy.
European journal of cell biology
Rosencrans WM,Walsh ZH,Houerbi N,Blum A,Belew M,Liu C,Chernak B,Brauer PR,Trazo A,Olson A,Hagos E
Wed Jan 01 00:00:00 UTC 2020
Globular CTRP9 protects cardiomyocytes from palmitic acid-induced oxidative stress by enhancing autophagic flux.
Chemico-biological interactions
Zuo A,Li J,Zhao X,Li T,Lei S,Chen J,Xu D,Song C,Li N,Ruan S,Lyu L,Guo Y
Fri Sep 25 00:00:00 UTC 2020
Autophagy in endothelial cells regulates their haematopoiesis-supporting ability.
EBioMedicine
Lyu ZS,Cao XN,Wen Q,Mo XD,Zhao HY,Chen YH,Wang Y,Chang YJ,Xu LP,Zhang XH,Kong Y,Huang XJ
Sun Mar 01 00:00:00 UTC 2020
Acupuncture protects against cerebral ischemia-reperfusion injury via suppressing endoplasmic reticulum stress-mediated autophagy and apoptosis.
Molecular medicine (Cambridge, Mass.)
Sun X,Liu H,Sun Z,Zhang B,Wang X,Liu T,Pan T,Gao Y,Jiang X,Li H
Tue Nov 10 00:00:00 UTC 2020
Acupuncture protects against cerebral ischemia-reperfusion injury via suppressing endoplasmic reticulum stress-mediated autophagy and apoptosis.
Molecular medicine (Cambridge, Mass.)
Sun X,Liu H,Sun Z,Zhang B,Wang X,Liu T,Pan T,Gao Y,Jiang X,Li H
Tue Nov 10 00:00:00 UTC 2020
The Effect of Autophagic Activity on the Function of Apheresis Platelets and on the Efficacy of Clinical Platelet Transfusion.
Transfusion medicine and hemotherapy : offizielles Organ der Deutschen Gesellschaft fur Transfusionsmedizin und Immunhamatologie
Tang H,Gao M,Fu Y,Gui R,Ma X
Wed Jul 01 00:00:00 UTC 2020
The Effect of Autophagic Activity on the Function of Apheresis Platelets and on the Efficacy of Clinical Platelet Transfusion.
Transfusion medicine and hemotherapy : offizielles Organ der Deutschen Gesellschaft fur Transfusionsmedizin und Immunhamatologie
Tang H,Gao M,Fu Y,Gui R,Ma X
Wed Jul 01 00:00:00 UTC 2020
GL-V9 reverses adriamycin resistance in hepatocellular carcinoma cells by affecting JNK2-related autophagy.
Chinese journal of natural medicines
Yang Y,Liao Y,Gui YP,Zhao L,Guo LB
Wed Jul 01 00:00:00 UTC 2020
The Class III PI3K/Beclin-1 Autophagic Pathway Participates in the mmLDL-Induced Upregulation of ETA Receptor in Mouse Mesenteric Arteries.
Advances in pharmacological and pharmaceutical sciences
Xie X,Chen C,Xu CB,Lin J,Cao L,Chen G,Li J
Mon Sep 28 00:00:00 UTC 2020
Bergenin Exerts Hepatoprotective Effects by Inhibiting the Release of Inflammatory Factors, Apoptosis and Autophagy via the PPAR-γ Pathway.
Drug design, development and therapy
Xiang S,Chen K,Xu L,Wang T,Guo C
Tue Dec 22 00:00:00 UTC 2020
Astragalus polysaccharide exerts anti-Parkinson via activating the PI3K/AKT/mTOR pathway to increase cellular autophagy level in vitro.
International journal of biological macromolecules
Tan Y,Yin L,Sun Z,Shao S,Chen W,Man X,Du Y,Chen Y
Mon Jun 15 00:00:00 UTC 2020
Potential therapeutic role of fibroblast growth factor 21 in neurodegeneration: Evidence for ameliorating parkinsonism via silent information regulator 2 homolog 1 and implication for gene therapy.
Neuropharmacology
Chen Y,Shen J,Qi G,Zha Q,Zhang C,Yao W,Gao X,Chen S
Tue Dec 15 00:00:00 UTC 2020
Potential therapeutic role of fibroblast growth factor 21 in neurodegeneration: Evidence for ameliorating parkinsonism via silent information regulator 2 homolog 1 and implication for gene therapy.
Neuropharmacology
Chen Y,Shen J,Qi G,Zha Q,Zhang C,Yao W,Gao X,Chen S
Tue Dec 15 00:00:00 UTC 2020
Nicotinamide Riboside Enhances Mitochondrial Proteostasis and Adult Neurogenesis through Activation of Mitochondrial Unfolded Protein Response Signaling in the Brain of ALS SOD1G93A Mice.
International journal of biological sciences
Zhou Q,Zhu L,Qiu W,Liu Y,Yang F,Chen W,Xu R
Mon Nov 02 00:00:00 UTC 2020
Spermidine improves gut barrier integrity and gut microbiota function in diet-induced obese mice.
Gut microbes
Ma L,Ni Y,Wang Z,Tu W,Ni L,Zhuge F,Zheng A,Hu L,Zhao Y,Zheng L,Fu Z
Mon Nov 09 00:00:00 UTC 2020
Cafestol preconditioning attenuates apoptosis and autophagy during hepatic ischemia-reperfusion injury by inhibiting ERK/PPARγ pathway.
International immunopharmacology
Ji J,Wu L,Feng J,Mo W,Wu J,Yu Q,Li S,Zhang J,Dai W,Xu X,Mao Y,Xu S,Chen K,Li J,Guo C
Wed Jul 01 00:00:00 UTC 2020
Prucalopride inhibits lung cancer cell proliferation, invasion, and migration through blocking of the PI3K/AKT/mTor signaling pathway.
Human & experimental toxicology
Chen M,Zhu LL,Su JL,Li GL,Wang J,Zhang YN
Sat Feb 01 00:00:00 UTC 2020
Discoidin Domain Receptors 1 Inhibition Alleviates Osteoarthritis via Enhancing Autophagy.
International journal of molecular sciences
Chou HC,Chen CH,Chou LY,Cheng TL,Kang L,Chuang SC,Lin YS,Ho ML,Wang YH,Lin SY,Wang CZ
Wed Sep 23 00:00:00 UTC 2020
Autophagy plays a protective role during Pseudomonas aeruginosa-induced apoptosis via ROS-MAPK pathway.
Innate immunity
Han L,Ma Q,Yu J,Gong Z,Ma C,Xu Y,Deng G,Wu X
Thu Oct 01 00:00:00 UTC 2020
Autophagy plays a protective role during Pseudomonas aeruginosa-induced apoptosis via ROS-MAPK pathway.
Innate immunity
Han L,Ma Q,Yu J,Gong Z,Ma C,Xu Y,Deng G,Wu X
Thu Oct 01 00:00:00 UTC 2020
A steroidal saponin isolated from Allium chinense simultaneously induces apoptosis and autophagy by modulating the PI3K/Akt/mTOR signaling pathway in human gastric adenocarcinoma.
Steroids
Xu J,Zhang M,Lin X,Wang Y,He X
Tue Sep 01 00:00:00 UTC 2020
A steroidal saponin isolated from Allium chinense simultaneously induces apoptosis and autophagy by modulating the PI3K/Akt/mTOR signaling pathway in human gastric adenocarcinoma.
Steroids
Xu J,Zhang M,Lin X,Wang Y,He X
Tue Sep 01 00:00:00 UTC 2020
Metformin Activates the AMPK-mTOR Pathway by Modulating lncRNA TUG1 to Induce Autophagy and Inhibit Atherosclerosis.
Drug design, development and therapy
You G,Long X,Song F,Huang J,Tian M,Xiao Y,Deng S,Wu Q
Tue Jan 05 00:00:00 UTC 2021
Metformin Activates the AMPK-mTOR Pathway by Modulating lncRNA TUG1 to Induce Autophagy and Inhibit Atherosclerosis.
Drug design, development and therapy
You G,Long X,Song F,Huang J,Tian M,Xiao Y,Deng S,Wu Q
Tue Jan 05 00:00:00 UTC 2021
Electroacupuncture ameliorates cerebral ischemia/reperfusion injury by suppressing autophagy via the SIRT1-FOXO1 signaling pathway.
Aging
Mei ZG,Huang YG,Feng ZT,Luo YN,Yang SB,Du LP,Jiang K,Liu XL,Fu XY,Deng YH,Zhou HJ
Fri Jul 03 00:00:00 UTC 2020
Electroacupuncture ameliorates cerebral ischemia/reperfusion injury by suppressing autophagy via the SIRT1-FOXO1 signaling pathway.
Aging
Mei ZG,Huang YG,Feng ZT,Luo YN,Yang SB,Du LP,Jiang K,Liu XL,Fu XY,Deng YH,Zhou HJ
Fri Jul 03 00:00:00 UTC 2020
Melatonin alleviates defects induced by zearalenone during porcine embryo development.
Theriogenology
Yao X,Jiang H,Gao Q,Li YH,Xu YN,Kim NH
Wed Jul 15 00:00:00 UTC 2020
Baicalin Inhibits Coxsackievirus B3 Replication by Reducing Cellular Lipid Synthesis.
The American journal of Chinese medicine
Wang MJ,Yang CH,Jin Y,Wan CB,Qian WH,Xing F,Li X,Liu YY
Fri Sep 04 00:00:00 UTC 2020
Autophagy Promotes Duck Tembusu Virus Replication by Suppressing p62/SQSTM1-Mediated Innate Immune Responses In Vitro.
Vaccines
Hu Z,Pan Y,Cheng A,Zhang X,Wang M,Chen S,Zhu D,Liu M,Yang Q,Wu Y,Zhao X,Huang J,Zhang S,Mao S,Ou X,Yu Y,Zhang L,Liu Y,Tian B,Pan L,Rehman MU,Yin Z,Jia R
Mon Jan 13 00:00:00 UTC 2020
Autophagy Promotes Duck Tembusu Virus Replication by Suppressing p62/SQSTM1-Mediated Innate Immune Responses In Vitro.
Vaccines
Hu Z,Pan Y,Cheng A,Zhang X,Wang M,Chen S,Zhu D,Liu M,Yang Q,Wu Y,Zhao X,Huang J,Zhang S,Mao S,Ou X,Yu Y,Zhang L,Liu Y,Tian B,Pan L,Rehman MU,Yin Z,Jia R
Mon Jan 13 00:00:00 UTC 2020
Lanosterol modulates proteostasis via dissolving cytosolic sequestosomes/aggresome-like induced structures.
Biochimica et biophysica acta. Molecular cell research
Hu LD,Wang J,Chen XJ,Yan YB
Sat Feb 01 00:00:00 UTC 2020
Anti-insulin resistance effects of salidroside through mitochondrial quality control.
The Journal of endocrinology
You B,Dun Y,Zhang W,Jiang L,Li H,Xie M,Liu Y,Liu S
Sat Feb 01 00:00:00 UTC 2020
Effect of autophagy on irradiation‑induced damage in osteoblast‑like MC3T3‑E1 cells.
Molecular medicine reports
Li R,Yang W,Hu X,Zhou D,Huang K,Wang C,Li Y,Liu B
Thu Oct 01 00:00:00 UTC 2020
Long Noncoding RNA TUG1 Promotes Autophagy-Associated Paclitaxel Resistance by Sponging miR-29b-3p in Ovarian Cancer Cells.
OncoTargets and therapy
Gu L,Li Q,Liu H,Lu X,Zhu M
Wed Apr 13 00:00:00 UTC 2022
Pseudolaric acid B induces apoptosis in human rhabdomyosarcoma RD cells.
Oncology letters
Liu C,Wang F,Wang B,Wu T,Wang Y,Huo W,Zhang S,Su Y,Liu J,Liu Y,Yu J
Tue Dec 01 00:00:00 UTC 2020
Aberrant miR-1246 expression promotes radioresistance in non-small cell lung cancer: a potential prognostic biomarker and radiotherapy sensitization target.
American journal of cancer research
Fan L,Wang J,Cao Q,Ding X,Li B
Mon Nov 13 00:00:00 UTC 2023
4-Acetylantroquinonol B Inhibits Osteoclastogenesis by Inhibiting the Autophagy Pathway in a Simulated Microgravity Model.
International journal of molecular sciences
Wu CH,Ou CH,Yen IC,Lee SY
Tue Sep 22 00:00:00 UTC 2020
Diphyllin Improves High-Fat Diet-Induced Obesity in Mice Through Brown and Beige Adipocytes.
Frontiers in endocrinology
Duan YN,Ge X,Jiang HW,Zhang HJ,Zhao Y,Li JL,Zhang W,Li JY
Fri May 21 00:00:00 UTC 2021
Evaluation of toxicity of halloysite nanotubes and multi-walled carbon nanotubes to endothelial cells in vitro and blood vessels in vivo.
Nanotoxicology
Wu B,Jiang M,Liu X,Huang C,Gu Z,Cao Y
Thu Oct 01 00:00:00 UTC 2020
The vicious cycle between ferritinophagy and ROS production triggered EMT inhibition of gastric cancer cells was through p53/AKT/mTor pathway.
Chemico-biological interactions
Xu Z,Feng J,Li Y,Guan D,Chen H,Zhai X,Zhang L,Li C,Li C
Tue Sep 01 00:00:00 UTC 2020
14-3-3ζ binds to and stabilizes phospho-beclin 1S295 and induces autophagy in hepatocellular carcinoma cells.
Journal of cellular and molecular medicine
Tang Y,Zhang Y,Liu S,Sun Z,Wang C,Li L,Zhou W,Cheng S
Wed Jan 01 00:00:00 UTC 2020
A lysosome-targeted ruthenium(II) polypyridyl complex as photodynamic anticancer agent.
Journal of inorganic biochemistry
Chen J,Tao Q,Wu J,Wang M,Su Z,Qian Y,Yu T,Wang Y,Xue X,Liu HK
Tue Sep 01 00:00:00 UTC 2020
Mainstream cigarette smoke induces autophagy and promotes apoptosis in oral mucosal epithelial cells.
Archives of oral biology
Liu Q,Zhao M,Chen W,Xu K,Huang F,Qu J,Xu Z,Wang X,Wang Y,Zhu Y,Wang W
Sun Mar 01 00:00:00 UTC 2020
Mainstream cigarette smoke induces autophagy and promotes apoptosis in oral mucosal epithelial cells.
Archives of oral biology
Liu Q,Zhao M,Chen W,Xu K,Huang F,Qu J,Xu Z,Wang X,Wang Y,Zhu Y,Wang W
Sun Mar 01 00:00:00 UTC 2020
Mainstream cigarette smoke induces autophagy and promotes apoptosis in oral mucosal epithelial cells.
Archives of oral biology
Liu Q,Zhao M,Chen W,Xu K,Huang F,Qu J,Xu Z,Wang X,Wang Y,Zhu Y,Wang W
Sun Mar 01 00:00:00 UTC 2020
Mainstream cigarette smoke induces autophagy and promotes apoptosis in oral mucosal epithelial cells.
Archives of oral biology
Liu Q,Zhao M,Chen W,Xu K,Huang F,Qu J,Xu Z,Wang X,Wang Y,Zhu Y,Wang W
Sun Mar 01 00:00:00 UTC 2020
Mainstream cigarette smoke induces autophagy and promotes apoptosis in oral mucosal epithelial cells.
Archives of oral biology
Liu Q,Zhao M,Chen W,Xu K,Huang F,Qu J,Xu Z,Wang X,Wang Y,Zhu Y,Wang W
Sun Mar 01 00:00:00 UTC 2020
Mainstream cigarette smoke induces autophagy and promotes apoptosis in oral mucosal epithelial cells.
Archives of oral biology
Liu Q,Zhao M,Chen W,Xu K,Huang F,Qu J,Xu Z,Wang X,Wang Y,Zhu Y,Wang W
Sun Mar 01 00:00:00 UTC 2020
Long Noncoding RNA UCA1 Is Related to Autophagy and Apoptosis in Endometrial Stromal Cells.
Frontiers in oncology
Jiang L,Wan Y,Feng Z,Liu D,Ouyang L,Li Y,Liu K
Thu Apr 21 00:00:00 UTC 2022
Panax notoginseng saponins prevent senescence and inhibit apoptosis by regulating the PI3K‑AKT‑mTOR pathway in osteoarthritic chondrocytes.
International journal of molecular medicine
Zhang Y,Cai W,Han G,Zhou S,Li J,Chen M,Li H
Wed Apr 01 00:00:00 UTC 2020
Knockdown of TXNDC9 induces apoptosis and autophagy in glioma and mediates cell differentiation by p53 activation.
Aging
Zheng T,Chen K,Zhang X,Feng H,Shi Y,Liu L,Zhang J,Chen Y
Tue Sep 08 00:00:00 UTC 2020
DUSP5 (dual-specificity protein phosphatase 5) suppresses BCG-induced autophagy via ERK 1/2 signaling pathway.
Molecular immunology
Luo J,Xue D,Song F,Liu X,Li W,Wang Y
Thu Oct 01 00:00:00 UTC 2020
Baicalin Protects Human OA Chondrocytes Against IL-1β-Induced Apoptosis and ECM Degradation by Activating Autophagy via MiR-766-3p/AIFM1 Axis.
Drug design, development and therapy
Li Z,Cheng J,Liu J
Fri Jun 04 00:00:00 UTC 2021
Beclin1 Binds to Enterovirus 71 3D Protein to Promote the Virus Replication.
Viruses
Xiang Q,Wan P,Yang G,Huang S,Qin M,Yang H,Luo Z,Wu K,Wu J
Tue Jul 14 00:00:00 UTC 2020
MicroRNA-1276 Promotes Colon Cancer Cell Proliferation by Negatively Regulating LACTB.
Cancer management and research
Wang C,Shi Z,Hong Z,Pan J,Chen Z,Qiu C,Zhuang H,Zheng X
Mon Apr 18 00:00:00 UTC 2022
Morroniside Inhibits H2O2-Induced Podocyte Apoptosis by Down-Regulating NOX4 Expression Controlled by Autophagy In Vitro.
Frontiers in pharmacology
Gao X,Liu Y,Wang L,Sai N,Liu Y,Ni J
Thu Oct 22 00:00:00 UTC 2020
Morroniside Inhibits H2O2-Induced Podocyte Apoptosis by Down-Regulating NOX4 Expression Controlled by Autophagy In Vitro.
Frontiers in pharmacology
Gao X,Liu Y,Wang L,Sai N,Liu Y,Ni J
Thu Oct 22 00:00:00 UTC 2020
Empagliflozin Alleviates Hepatic Steatosis by Activating the AMPK-TET2-Autophagy Pathway in vivo and in vitro.
Frontiers in pharmacology
Li T,Fang T,Xu L,Liu X,Li X,Xue M,Yu X,Sun B,Chen L
Wed Feb 10 00:00:00 UTC 2021
DpdtC-Induced EMT Inhibition in MGC-803 Cells Was Partly through Ferritinophagy-Mediated ROS/p53 Pathway.
Oxidative medicine and cellular longevity
Feng J,Li C,Xu R,Li Y,Hou Q,Feng R,Wang S,Zhang L,Li C
Wed Dec 16 00:00:00 UTC 2020
Amino Acids Regulate Cisplatin Insensitivity in Neuroblastoma.
Cancers
Gunda V,Pathania AS,Chava S,Prathipati P,Chaturvedi NK,Coulter DW,Pandey MK,Durden DL,Challagundla KB
Thu Sep 10 00:00:00 UTC 2020
LL-37-Induced Autophagy Contributed to the Elimination of Live Porphyromonas gingivalis Internalized in Keratinocytes.
Frontiers in cellular and infection microbiology
Yang X,Niu L,Pan Y,Feng X,Liu J,Guo Y,Pan C,Geng F,Tang X
Thu Jun 17 00:00:00 UTC 2021
NeuroHeal Treatment Alleviates Neuropathic Pain and Enhances Sensory Axon Regeneration.
Cells
Romeo-Guitart D,Casas C
Fri Mar 27 00:00:00 UTC 2020
LETM1 Knockdown Promotes Autophagy and Apoptosis Through AMP-Activated Protein Kinase Phosphorylation-Mediated Beclin-1/Bcl-2 Complex Dissociation in Hepatocellular Carcinoma.
Frontiers in oncology
Zhou B,Yang C,Yan X,Shi Z,Xiao H,Wei X,Jiang N,Wu Z
Wed Feb 10 00:00:00 UTC 2021
LETM1 Knockdown Promotes Autophagy and Apoptosis Through AMP-Activated Protein Kinase Phosphorylation-Mediated Beclin-1/Bcl-2 Complex Dissociation in Hepatocellular Carcinoma.
Frontiers in oncology
Zhou B,Yang C,Yan X,Shi Z,Xiao H,Wei X,Jiang N,Wu Z
Wed Feb 10 00:00:00 UTC 2021
Intra-Articular Injection of (-)-Epigallocatechin 3-Gallate to Attenuate Articular Cartilage Degeneration by Enhancing Autophagy in a Post-Traumatic Osteoarthritis Rat Model.
Antioxidants (Basel, Switzerland)
Huang HT,Cheng TL,Ho CJ,Huang HH,Lu CC,Chuang SC,Li JY,Lee TC,Chen ST,Lin YS,Lee CY,Kang L,Lin SY,Chen CH
Wed Dec 23 00:00:00 UTC 2020
Eugenol Attenuates Cerebral Ischemia-Reperfusion Injury by Enhancing Autophagy via AMPK-mTOR-P70S6K Pathway.
Frontiers in pharmacology
Sun X,Wang D,Zhang T,Lu X,Duan F,Ju L,Zhuang X,Jiang X
Mon Sep 28 00:00:00 UTC 2020
Eugenol Attenuates Cerebral Ischemia-Reperfusion Injury by Enhancing Autophagy via AMPK-mTOR-P70S6K Pathway.
Frontiers in pharmacology
Sun X,Wang D,Zhang T,Lu X,Duan F,Ju L,Zhuang X,Jiang X
Mon Sep 28 00:00:00 UTC 2020
Inhibiting PKCβ2 protects HK-2 cells against meglumine diatrizoate and AGEs-induced apoptosis and autophagy.
Annals of translational medicine
Jiang W,Zhao W,Ye F,Huang S,Wu Y,Chen H,Zhou R,Fu G
Sun Mar 01 00:00:00 UTC 2020
Olaquindox-Induced Liver Damage Involved the Crosstalk of Oxidative Stress and p53 In Vivo and In Vitro.
Oxidative medicine and cellular longevity
Li D,Pei X,Qin X,Liu X,Li C,Li L,Dai C,Xiao X,Tang S
Thu Sep 02 00:00:00 UTC 2021
Morusin induces apoptosis and autophagy via JNK, ERK and PI3K/Akt signaling in human lung carcinoma cells.
Chemico-biological interactions
Wang J,Liu X,Zheng H,Liu Q,Zhang H,Wang X,Shen T,Wang S,Ren D
Sun Nov 01 00:00:00 UTC 2020
SUMOylation of Pdia3 exacerbates proinsulin misfolding and ER stress in pancreatic beta cells.
Journal of molecular medicine (Berlin, Germany)
Li N,Luo X,Yu Q,Yang P,Chen Z,Wang X,Jiang J,Xu J,Gong Q,Eizirik DL,Zhou Z,Zhao J,Xiong F,Zhang S,Wang CY
Tue Dec 01 00:00:00 UTC 2020
The Acetone Indigo Red Dehydrating Agent IF203 Induces HepG2 Cell Death Through Cell Cycle Arrest, Autophagy and Apoptosis.
OncoTargets and therapy
Shang Y,Wang Q,Li J,Zhao Q,Huang X,Dong H,Liu H,Zhang Y,Zhang J,Gui R,Nie X
Tue Apr 12 00:00:00 UTC 2022
The renoprotective effect of diosgenin on aristolochic acid I-induced renal injury in rats: impact on apoptosis, mitochondrial dynamics and autophagy.
Food & function
Jin C,Miao X,Zhong Y,Han J,Liu Q,Zhu J,Xia X,Peng X
Wed Sep 23 00:00:00 UTC 2020
Lead exposure induces cell autophagy via blocking the Akt/mTOR signaling in rat astrocytes.
The Journal of toxicological sciences
Huang Y,Liao Y,Zhang H,Li S
Fri Oct 09 00:00:00 UTC 2020
Astragaloside IV prevents myocardial hypertrophy induced by mechanical stress by activating autophagy and reducing inflammation.
American journal of translational research
Zhang T,Wang H,Lu M,Zhao K,Yin J,Liu Y,Sun Y
Tue Oct 13 00:00:00 UTC 2020
The Preparation of Ginsenoside Rg5, Its Antitumor Activity against Breast Cancer Cells and Its Targeting of PI3K.
Nutrients
Liu Y,Fan D
Sat Jan 18 00:00:00 UTC 2020
The Preparation of Ginsenoside Rg5, Its Antitumor Activity against Breast Cancer Cells and Its Targeting of PI3K.
Nutrients
Liu Y,Fan D
Sat Jan 18 00:00:00 UTC 2020
Exosomes derived from mesenchymal stem cells repair a Parkinson's disease model by inducing autophagy.
Cell death & disease
Chen HX,Liang FC,Gu P,Xu BL,Xu HJ,Wang WT,Hou JY,Xie DX,Chai XQ,An SJ
Mon Apr 27 00:00:00 UTC 2020
Anticancer Effects of Honokiol via Mitochondrial Dysfunction Are Strongly Enhanced by the Mitochondria-Targeting Carrier Berberine.
Journal of medicinal chemistry
Shi X,Zhang T,Lou H,Song H,Li C,Fan P
Thu Oct 22 00:00:00 UTC 2020
N‑linoleyltyrosine protects PC12 cells against oxidative damage via autophagy: Possible involvement of CB1 receptor regulation.
International journal of molecular medicine
Liu X,Wu Y,Zhou D,Xie Y,Zhou Y,Lu Y,Yang R,Liu S
Sun Nov 01 00:00:00 UTC 2020
N‑linoleyltyrosine protects PC12 cells against oxidative damage via autophagy: Possible involvement of CB1 receptor regulation.
International journal of molecular medicine
Liu X,Wu Y,Zhou D,Xie Y,Zhou Y,Lu Y,Yang R,Liu S
Sun Nov 01 00:00:00 UTC 2020
Cellular evidence of autophagy in Sertoli cells during spermatogenesis in goats.
Theriogenology
Qu W,Tarique I,Deng B,Zhang Y,Haseeb A,Chen Q,Yang P
Tue Sep 15 00:00:00 UTC 2020
Autophagy Contributes to Host Immunity and Protection against Zika Virus Infection via Type I IFN Signaling.
Mediators of inflammation
Huang Y,Wang Y,Meng S,Chen Z,Kong H,Pan T,Lu G,Li X
Thu Jul 08 00:00:00 UTC 2021
Autophagy Contributes to Host Immunity and Protection against Zika Virus Infection via Type I IFN Signaling.
Mediators of inflammation
Huang Y,Wang Y,Meng S,Chen Z,Kong H,Pan T,Lu G,Li X
Thu Jul 08 00:00:00 UTC 2021
Upregulation of Parkin Accelerates Osteoblastic Differentiation of Bone Marrow-Derived Mesenchymal Stem Cells and Bone Regeneration by Enhancing Autophagy and β-Catenin Signaling.
Frontiers in cell and developmental biology
Zhang W,Hou W,Chen M,Chen E,Xue D,Ye C,Li W,Pan Z
Tue Oct 13 00:00:00 UTC 2020
Effect of combination of curcumin and GLUT-1 AS-ODN on radiosensitivity of laryngeal carcinoma through regulating autophagy.
Head & neck
Dai LB,Yu Q,Zhou SH,Bao YY,Zhong JT,Shen LF,Lu ZJ,Fan J,Huang YP
Tue Sep 01 00:00:00 UTC 2020
Autophagy and Hsp70 activation alleviate oral epithelial cell death induced by food-derived hypertonicity.
Cell stress & chaperones
Yang J,Zhang H,Sun S,Wang X,Guan Y,Mi Q,Zeng W,Xiang H,Zhu H,Zou X,You Y,Xiang Y,Gao Q
Sun Mar 01 00:00:00 UTC 2020
Enterovirus 71 induces neural cell apoptosis and autophagy through promoting ACOX1 downregulation and ROS generation.
Virulence
You L,Chen J,Liu W,Xiang Q,Luo Z,Wang W,Xu W,Wu K,Zhang Q,Liu Y,Wu J
Tue Dec 01 00:00:00 UTC 2020
Alantolactone inhibits cell autophagy and promotes apoptosis via AP2M1 in acute lymphoblastic leukemia.
Cancer cell international
Shi C,Lan W,Wang Z,Yang D,Wei J,Liu Z,Teng Y,Gu M,Yuan T,Cao F,Zhou J,Li Y
Sat Apr 16 00:00:00 UTC 2022
Aldehyde dehydrogenase 2 regulates autophagy via the Akt-mTOR pathway to mitigate renal ischemia-reperfusion injury in hypothermic machine perfusion.
Life sciences
Lin D,Xiang T,Qiu Q,Leung J,Xu J,Zhou W,Hu Q,Lan J,Liu Z,Zhong Z,Wang Y,Ye Q
Wed Jul 15 00:00:00 UTC 2020
Arsenic induces dysfunctional autophagy via dual regulation of mTOR pathway and Beclin1-Vps34/PI3K complex in MLTC-1 cells.
Journal of hazardous materials
Liang C,Feng Z,Manthari RK,Wang C,Han Y,Fu W,Wang J,Zhang J
Fri Jun 05 00:00:00 UTC 2020
Drp1-associated mitochondrial dysfunction and mitochondrial autophagy: a novel mechanism in triptolide-induced hepatotoxicity.
Cell biology and toxicology
Hasnat M,Yuan Z,Naveed M,Khan A,Raza F,Xu D,Ullah A,Sun L,Zhang L,Jiang Z
Sat Jun 01 00:00:00 UTC 2019
Drp1-associated mitochondrial dysfunction and mitochondrial autophagy: a novel mechanism in triptolide-induced hepatotoxicity.
Cell biology and toxicology
Hasnat M,Yuan Z,Naveed M,Khan A,Raza F,Xu D,Ullah A,Sun L,Zhang L,Jiang Z
Sat Jun 01 00:00:00 UTC 2019
Drp1-associated mitochondrial dysfunction and mitochondrial autophagy: a novel mechanism in triptolide-induced hepatotoxicity.
Cell biology and toxicology
Hasnat M,Yuan Z,Naveed M,Khan A,Raza F,Xu D,Ullah A,Sun L,Zhang L,Jiang Z
Sat Jun 01 00:00:00 UTC 2019
Drp1-associated mitochondrial dysfunction and mitochondrial autophagy: a novel mechanism in triptolide-induced hepatotoxicity.
Cell biology and toxicology
Hasnat M,Yuan Z,Naveed M,Khan A,Raza F,Xu D,Ullah A,Sun L,Zhang L,Jiang Z
Sat Jun 01 00:00:00 UTC 2019
Melatonin protects against sepsis-induced cardiac dysfunction by regulating apoptosis and autophagy via activation of SIRT1 in mice.
Life sciences
Zhang WX,He BM,Wu Y,Qiao JF,Peng ZY
Tue Jan 15 00:00:00 UTC 2019
Melatonin protects against sepsis-induced cardiac dysfunction by regulating apoptosis and autophagy via activation of SIRT1 in mice.
Life sciences
Zhang WX,He BM,Wu Y,Qiao JF,Peng ZY
Tue Jan 15 00:00:00 UTC 2019
Roles of mitochondrial fission inhibition in developmental fluoride neurotoxicity: mechanisms of action in vitro and associations with cognition in rats and children.
Archives of toxicology
Zhao Q,Niu Q,Chen J,Xia T,Zhou G,Li P,Dong L,Xu C,Tian Z,Luo C,Liu L,Zhang S,Wang A
Fri Mar 01 00:00:00 UTC 2019
Roles of mitochondrial fission inhibition in developmental fluoride neurotoxicity: mechanisms of action in vitro and associations with cognition in rats and children.
Archives of toxicology
Zhao Q,Niu Q,Chen J,Xia T,Zhou G,Li P,Dong L,Xu C,Tian Z,Luo C,Liu L,Zhang S,Wang A
Fri Mar 01 00:00:00 UTC 2019
AZD9291 promotes autophagy and inhibits PI3K/Akt pathway in NSCLC cancer cells.
Journal of cellular biochemistry
Zhang Z,Zhang M,Liu H,Yin W
Tue Jan 01 00:00:00 UTC 2019
AZD9291 promotes autophagy and inhibits PI3K/Akt pathway in NSCLC cancer cells.
Journal of cellular biochemistry
Zhang Z,Zhang M,Liu H,Yin W
Tue Jan 01 00:00:00 UTC 2019
AZD9291 promotes autophagy and inhibits PI3K/Akt pathway in NSCLC cancer cells.
Journal of cellular biochemistry
Zhang Z,Zhang M,Liu H,Yin W
Tue Jan 01 00:00:00 UTC 2019
AZD9291 promotes autophagy and inhibits PI3K/Akt pathway in NSCLC cancer cells.
Journal of cellular biochemistry
Zhang Z,Zhang M,Liu H,Yin W
Tue Jan 01 00:00:00 UTC 2019
Alleviation of hepatic fibrosis and autophagy via inhibition of transforming growth factor-β1/Smads pathway through shikonin.
Journal of gastroenterology and hepatology
Liu T,Xu L,Wang C,Chen K,Xia Y,Li J,Li S,Wu L,Feng J,Xu S,Wang W,Lu X,Fan X,Mo W,Zhou Y,Zhao Y,Guo C
Tue Jan 01 00:00:00 UTC 2019
Alleviation of hepatic fibrosis and autophagy via inhibition of transforming growth factor-β1/Smads pathway through shikonin.
Journal of gastroenterology and hepatology
Liu T,Xu L,Wang C,Chen K,Xia Y,Li J,Li S,Wu L,Feng J,Xu S,Wang W,Lu X,Fan X,Mo W,Zhou Y,Zhao Y,Guo C
Tue Jan 01 00:00:00 UTC 2019
The synergistic effect of NOD2 and TLR4 on the activation of autophagy in human submandibular gland inflammation.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Li J,Li B,Cheng Y,Meng Q,Wei L,Li W,Zhang J,Huang S
Tue Jan 01 00:00:00 UTC 2019
The synergistic effect of NOD2 and TLR4 on the activation of autophagy in human submandibular gland inflammation.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Li J,Li B,Cheng Y,Meng Q,Wei L,Li W,Zhang J,Huang S
Tue Jan 01 00:00:00 UTC 2019
The synergistic effect of NOD2 and TLR4 on the activation of autophagy in human submandibular gland inflammation.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Li J,Li B,Cheng Y,Meng Q,Wei L,Li W,Zhang J,Huang S
Tue Jan 01 00:00:00 UTC 2019
The synergistic effect of NOD2 and TLR4 on the activation of autophagy in human submandibular gland inflammation.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Li J,Li B,Cheng Y,Meng Q,Wei L,Li W,Zhang J,Huang S
Tue Jan 01 00:00:00 UTC 2019
Platelet-Membrane-Camouflaged Black Phosphorus Quantum Dots Enhance Anticancer Effect Mediated by Apoptosis and Autophagy.
ACS applied materials & interfaces
Shang Y,Wang Q,Wu B,Zhao Q,Li J,Huang X,Chen W,Gui R
Wed Aug 07 00:00:00 UTC 2019
Potent effects of dioscin against hepatocellular carcinoma through regulating TP53-induced glycolysis and apoptosis regulator (TIGAR)-mediated apoptosis, autophagy, and DNA damage.
British journal of pharmacology
Mao Z,Han X,Chen D,Xu Y,Xu L,Yin L,Sun H,Qi Y,Fang L,Liu K,Peng J
Mon Apr 01 00:00:00 UTC 2019
Pterostilbene, An Active Constituent of Blueberries, Suppresses Proliferation Potential of Human Cholangiocarcinoma via Enhancing the Autophagic Flux.
Frontiers in pharmacology
Wang D,Guo H,Yang H,Wang D,Gao P,Wei W
Tue Sep 29 00:00:00 UTC 2020
The ALS-FTD-linked gene product, C9orf72, regulates neuronal morphogenesis via autophagy.
Autophagy
Ho WY,Tai YK,Chang JC,Liang J,Tyan SH,Chen S,Guan JL,Zhou H,Shen HM,Koo E,Ling SC
Wed May 01 00:00:00 UTC 2019
HMGB1/TLR4 promotes apoptosis and reduces autophagy of hippocampal neurons in diabetes combined with OSA.
Life sciences
Guo X,Shi Y,Du P,Wang J,Han Y,Sun B,Feng J
Sun Dec 15 00:00:00 UTC 2019
HMGB1/TLR4 promotes apoptosis and reduces autophagy of hippocampal neurons in diabetes combined with OSA.
Life sciences
Guo X,Shi Y,Du P,Wang J,Han Y,Sun B,Feng J
Sun Dec 15 00:00:00 UTC 2019
Prevention of Huntington's Disease-Like Behavioral Deficits in R6/1 Mouse by Tolfenamic Acid Is Associated with Decreases in Mutant Huntingtin and Oxidative Stress.
Oxidative medicine and cellular longevity
Liu P,Li Y,Yang W,Liu D,Ji X,Chi T,Guo Z,Li L,Zou L
Wed Aug 28 00:00:00 UTC 2019
Nonstructural Protein NP1 of Human Bocavirus 1 suppresses the growth of A549 cell by promoting autophagy.
The new microbiologica
Zhu J,Li J,Wang X,Feng X,Li Y
Mon Jul 01 00:00:00 UTC 2019
Effects of long-term fluoride exposure on cognitive ability and the underlying mechanisms: Role of autophagy and its association with apoptosis.
Toxicology and applied pharmacology
Zhou G,Tang S,Yang L,Niu Q,Chen J,Xia T,Wang S,Wang M,Zhao Q,Liu L,Li P,Dong L,Yang K,Zhang S,Wang A
Sun Sep 01 00:00:00 UTC 2019
Effects of long-term fluoride exposure on cognitive ability and the underlying mechanisms: Role of autophagy and its association with apoptosis.
Toxicology and applied pharmacology
Zhou G,Tang S,Yang L,Niu Q,Chen J,Xia T,Wang S,Wang M,Zhao Q,Liu L,Li P,Dong L,Yang K,Zhang S,Wang A
Sun Sep 01 00:00:00 UTC 2019
Autophagy impairment contributes to PBDE-47-induced developmental neurotoxicity and its relationship with apoptosis.
Theranostics
Li P,Ma R,Dong L,Liu L,Zhou G,Tian Z,Zhao Q,Xia T,Zhang S,Wang A
Fri Aug 14 00:00:00 UTC 2020
Autophagy impairment contributes to PBDE-47-induced developmental neurotoxicity and its relationship with apoptosis.
Theranostics
Li P,Ma R,Dong L,Liu L,Zhou G,Tian Z,Zhao Q,Xia T,Zhang S,Wang A
Fri Aug 14 00:00:00 UTC 2020
Overexpression of iASPP is required for autophagy in response to oxidative stress in choriocarcinoma.
BMC cancer
Chan KK,Wong ES,Wong IT,Cheung CL,Wong OG,Ngan HY,Cheung AN
Tue Oct 15 00:00:00 UTC 2019
Rapamycin attenuates articular cartilage degeneration by inhibiting β-catenin in a murine model of osteoarthritis.
Connective tissue research
Ma L,Liu Y,Zhao X,Li P,Jin Q
Sun Sep 01 00:00:00 UTC 2019
Microglia as modulators of exosomal alpha-synuclein transmission.
Cell death & disease
Xia Y,Zhang G,Han C,Ma K,Guo X,Wan F,Kou L,Yin S,Liu L,Huang J,Xiong N,Wang T
Wed Feb 20 00:00:00 UTC 2019
Effects of Fluoride on Autophagy in Mouse Sertoli Cells.
Biological trace element research
Feng Z,Liang C,Manthari RK,Wang C,Zhang J
Fri Feb 01 00:00:00 UTC 2019
The antidepressant effects of apigenin are associated with the promotion of autophagy via the mTOR/AMPK/ULK1 pathway.
Molecular medicine reports
Zhang X,Bu H,Jiang Y,Sun G,Jiang R,Huang X,Duan H,Huang Z,Wu Q
Sun Sep 01 00:00:00 UTC 2019
Inhibition of glycogen catabolism induces intrinsic apoptosis and augments multikinase inhibitors in hepatocellular carcinoma cells.
Experimental cell research
Barot S,Abo-Ali EM,Zhou DL,Palaguachi C,Dukhande VV
Thu Aug 15 00:00:00 UTC 2019
Inhibition of glycogen catabolism induces intrinsic apoptosis and augments multikinase inhibitors in hepatocellular carcinoma cells.
Experimental cell research
Barot S,Abo-Ali EM,Zhou DL,Palaguachi C,Dukhande VV
Thu Aug 15 00:00:00 UTC 2019
Enhancing autophagy by down-regulating GSK-3β alleviates cisplatin-induced ototoxicity in vivo and in vitro.
Toxicology letters
Liu T,Zong S,Luo P,Qu Y,Wen Y,Du P,Xiao H
Tue Oct 01 00:00:00 UTC 2019
Rafoxanide promotes apoptosis and autophagy of gastric cancer cells by suppressing PI3K /Akt/mTOR pathway.
Experimental cell research
Liu JZ,Hu YL,Feng Y,Guo YB,Liu YF,Yang JL,Mao QS,Xue WJ
Sun Dec 15 00:00:00 UTC 2019
Isodunnianol alleviates doxorubicin-induced myocardial injury by activating protective autophagy.
Food & function
Chen C,Jiang L,Zhang M,Pan X,Peng C,Huang W,Jiang Q
Wed May 22 00:00:00 UTC 2019
STAT3 phosphorylation mediates high glucose-impaired cell autophagy in an HDAC1-dependent and -independent manner in Schwann cells of diabetic peripheral neuropathy.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Du W,Wang N,Li F,Jia K,An J,Liu Y,Wang Y,Zhu L,Zhao S,Hao J
Mon Jul 01 00:00:00 UTC 2019
Conditioned medium obtained from human amniotic mesenchymal stem cells attenuates focal cerebral ischemia/reperfusion injury in rats by targeting mTOR pathway.
Journal of chemical neuroanatomy
Nazarinia D,Aboutaleb N,Gholamzadeh R,Nasseri Maleki S,Mokhtari B,Nikougoftar M
Sun Dec 01 00:00:00 UTC 2019
Berberine Maintains the Neutrophil N1 Phenotype to Reverse Cancer Cell Resistance to Doxorubicin.
Frontiers in pharmacology
Zhang S,Zhou L,Zhang M,Wang Y,Wang M,Du J,Gu W,Kui F,Li J,Geng S,Du G
Fri Dec 22 00:00:00 UTC 2023
MicroRNA-101 regulates autophagy, proliferation and apoptosis via targeting EZH2 in laryngeal squamous cell carcinoma.
Neoplasma
Chen L,Jia J,Zang Y,Li J,Wan B
Tue Jul 23 00:00:00 UTC 2019
The long non-coding RNA H19 induces hypoxia/reoxygenation injury by up-regulating autophagy in the hepatoma carcinoma cells.
Biological research
Cui C,Li Z,Wu D
Thu Jun 13 00:00:00 UTC 2019
Sophoridine Inhibits Human Colorectal Cancer Progression via Targeting MAPKAPK2.
Molecular cancer research : MCR
Wang R,Liu H,Shao Y,Wang K,Yin S,Qiu Y,Wu H,Liu E,Wang T,Gao X,Yu H
Sun Dec 01 00:00:00 UTC 2019
Excessive Methionine Supplementation Exacerbates the Development of Abdominal Aortic Aneurysm in Rats.
Journal of vascular research
Fan Y,Li N,Liu C,Dong H,Hu X
Mon Jan 27 00:00:00 UTC 2020
Corticosterone Excess-Mediated Mitochondrial Damage Induces Hippocampal Neuronal Autophagy in Mice Following Cold Exposure.
Animals : an open access journal from MDPI
Xu B,Lang L,Li S,Yuan J,Wang J,Yang H,Lian S
Sat Sep 14 00:00:00 UTC 2019
Isorhamnetin Inhibits Liver Fibrosis by Reducing Autophagy and Inhibiting Extracellular Matrix Formation via the TGF-β1/Smad3 and TGF-β1/p38 MAPK Pathways.
Mediators of inflammation
Liu N,Feng J,Lu X,Yao Z,Liu Q,Lv Y,Han Y,Deng J,Zhou Y
Mon Jan 27 00:00:00 UTC 2020
Effects of Exercise Training on the Autophagy-Related Muscular Proteins Expression in Ovariectomized Rats.
Frontiers in physiology
Zhong W,Shi X,Yuan H,Bu H,Wu L,Wang R
Wed Sep 30 00:00:00 UTC 2020
Transmembrane Protein 39A Promotes the Replication of Encephalomyocarditis Virus via Autophagy Pathway.
Frontiers in microbiology
Li X,Ma R,Li Q,Li S,Zhang H,Xie J,Bai J,Idris A,Feng R
Thu Oct 01 00:00:00 UTC 2020
Lithium targeting of AMPK protects against cisplatin-induced acute kidney injury by enhancing autophagy in renal proximal tubular epithelial cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Bao H,Zhang Q,Liu X,Song Y,Li X,Wang Z,Li C,Peng A,Gong R
Sun Dec 01 00:00:00 UTC 2019
Lithium targeting of AMPK protects against cisplatin-induced acute kidney injury by enhancing autophagy in renal proximal tubular epithelial cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Bao H,Zhang Q,Liu X,Song Y,Li X,Wang Z,Li C,Peng A,Gong R
Sun Dec 01 00:00:00 UTC 2019
Protective effects of UFL1 against endoplasmic reticulum stress-induced autophagy in bovine mammary epithelial cells.
Cell stress & chaperones
Kuang M,Li L,Li C,Wang G
Fri Nov 01 00:00:00 UTC 2019
Propofol Attenuates Hypoxia/Reoxygenation-Induced Apoptosis and Autophagy in HK-2 Cells by Inhibiting JNK Activation.
Yonsei medical journal
Wang H,Peng X,Huang Y,Xiao Y,Wang Z,Zhan L
Sun Dec 01 00:00:00 UTC 2019
Protective role of thymoquinone in sepsis-induced liver injury in BALB/c mice.
Experimental and therapeutic medicine
Wang F,Lei X,Zhao Y,Yu Q,Li Q,Zhao H,Pei Z
Sun Sep 01 00:00:00 UTC 2019
Inhibition of PIKfyve using YM201636 suppresses the growth of liver cancer via the induction of autophagy.
Oncology reports
Hou JZ,Xi ZQ,Niu J,Li W,Wang X,Liang C,Sun H,Fang D,Xie SQ
Fri Mar 01 00:00:00 UTC 2019
Fibroblast growth factor-21 alleviates hypoxia/reoxygenation injury in H9c2 cardiomyocytes by promoting autophagic flux.
International journal of molecular medicine
Ren Z,Xiao W,Zeng Y,Liu MH,Li GH,Tang ZH,Qu SL,Hao YM,Yuan HQ,Jiang ZS
Fri Mar 01 00:00:00 UTC 2019
Paeonol induces cytoprotective autophagy via blocking the Akt/mTOR pathway in ovarian cancer cells.
Cell death & disease
Gao L,Wang Z,Lu D,Huang J,Liu J,Hong L
Tue Aug 13 00:00:00 UTC 2019
Exogenous hydrogen sulfide mitigates NLRP3 inflammasome-mediated inflammation through promoting autophagy via the AMPK-mTOR pathway.
Biology open
Wang H,Zhong P,Sun L
Wed Jul 17 00:00:00 UTC 2019
Exogenous hydrogen sulfide mitigates NLRP3 inflammasome-mediated inflammation through promoting autophagy via the AMPK-mTOR pathway.
Biology open
Wang H,Zhong P,Sun L
Wed Jul 17 00:00:00 UTC 2019
Iron dysregulation in vascular dementia: Focused on the AMPK/autophagy pathway.
Brain research bulletin
Huo T,Jia Y,Yin C,Luo X,Zhao J,Wang Z,Lv P
Fri Nov 01 00:00:00 UTC 2019
Iron dysregulation in vascular dementia: Focused on the AMPK/autophagy pathway.
Brain research bulletin
Huo T,Jia Y,Yin C,Luo X,Zhao J,Wang Z,Lv P
Fri Nov 01 00:00:00 UTC 2019
Beclin 1, LC3, and p62 expression in paraquat-induced pulmonary fibrosis.
Human & experimental toxicology
Xu G,Wang X,Yu H,Wang C,Liu Y,Zhao R,Zhang G
Mon Jul 01 00:00:00 UTC 2019
Adiponectin inhibits high glucose-induced angiogenesis via inhibiting autophagy in RF/6A cells.
Journal of cellular physiology
Li R,Du J,Yao Y,Yao G,Wang X
Fri Nov 01 00:00:00 UTC 2019
C9orf72 intermediate repeats are associated with corticobasal degeneration, increased C9orf72 expression and disruption of autophagy.
Acta neuropathologica
Cali CP,Patino M,Tai YK,Ho WY,McLean CA,Morris CM,Seeley WW,Miller BL,Gaig C,Vonsattel JPG,White CL,Roeber S,Kretzschmar H,Troncoso JC,Troakes C,Gearing M,Ghetti B,Van Deerlin VM,Lee VM,Trojanowski JQ,Mok KY,Ling H,Dickson DW,Schellenberg GD,Ling SC,Lee EB
Fri Nov 01 00:00:00 UTC 2019
XIAOPI formula promotes breast cancer chemosensitivity via inhibiting CXCL1/HMGB1-mediated autophagy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang N,Yang B,Muhetaer G,Wang S,Zheng Y,Lu J,Li M,Zhang F,Situ H,Lin Y,Wang Z
Sun Dec 01 00:00:00 UTC 2019
XIAOPI formula promotes breast cancer chemosensitivity via inhibiting CXCL1/HMGB1-mediated autophagy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang N,Yang B,Muhetaer G,Wang S,Zheng Y,Lu J,Li M,Zhang F,Situ H,Lin Y,Wang Z
Sun Dec 01 00:00:00 UTC 2019
XIAOPI formula promotes breast cancer chemosensitivity via inhibiting CXCL1/HMGB1-mediated autophagy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang N,Yang B,Muhetaer G,Wang S,Zheng Y,Lu J,Li M,Zhang F,Situ H,Lin Y,Wang Z
Sun Dec 01 00:00:00 UTC 2019
XIAOPI formula promotes breast cancer chemosensitivity via inhibiting CXCL1/HMGB1-mediated autophagy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang N,Yang B,Muhetaer G,Wang S,Zheng Y,Lu J,Li M,Zhang F,Situ H,Lin Y,Wang Z
Sun Dec 01 00:00:00 UTC 2019
Visfatin Plays a Significant Role in Alleviating Lipopolysaccharide-Induced Apoptosis and Autophagy Through PI3K/AKT Signaling Pathway During Acute Lung Injury in Mice.
Archivum immunologiae et therapiae experimentalis
Wu XT,Ansari AR,Pang XX,Li HZ,Zhang ZW,Luo Y,Arshad M,Song H
Thu Aug 01 00:00:00 UTC 2019
Visfatin Plays a Significant Role in Alleviating Lipopolysaccharide-Induced Apoptosis and Autophagy Through PI3K/AKT Signaling Pathway During Acute Lung Injury in Mice.
Archivum immunologiae et therapiae experimentalis
Wu XT,Ansari AR,Pang XX,Li HZ,Zhang ZW,Luo Y,Arshad M,Song H
Thu Aug 01 00:00:00 UTC 2019
Grape Seed Proanthocyanidins Induce Autophagy and Modulate Survivin in HepG2 Cells and Inhibit Xenograft Tumor Growth in Vivo.
Nutrients
Wang L,Huang W,Zhan J
Fri Dec 06 00:00:00 UTC 2019
Rab18 Collaborates with Rab7 to Modulate Lysosomal and Autophagy Activities in the Nervous System: an Overlapping Mechanism for Warburg Micro Syndrome and Charcot-Marie-Tooth Neuropathy Type 2B.
Molecular neurobiology
Nian FS,Li LL,Cheng CY,Wu PC,Lin YT,Tang CY,Ren BS,Tai CY,Fann MJ,Kao LS,Hong CJ,Tsai JW
Sun Sep 01 00:00:00 UTC 2019
A novel ALK inhibitor ZYY inhibits Karpas299 cell growth in vitro and in a mouse xenograft model and induces protective autophagy.
Toxicology and applied pharmacology
Shen J,Wang J,Du J,Wang L,Zhou X,Chang X,Li Z,Zhai X,Zuo D,Wu Y
Fri Nov 15 00:00:00 UTC 2019
A novel ALK inhibitor ZYY inhibits Karpas299 cell growth in vitro and in a mouse xenograft model and induces protective autophagy.
Toxicology and applied pharmacology
Shen J,Wang J,Du J,Wang L,Zhou X,Chang X,Li Z,Zhai X,Zuo D,Wu Y
Fri Nov 15 00:00:00 UTC 2019
Protective effects of levo-tetrahydropalmatine on hepatic ischemia/reperfusion injury are mediated by inhibition of the ERK/NF-κB pathway.
International immunopharmacology
Yu Q,Wu L,Liu T,Li S,Feng J,Mao Y,Fan X,Guo C,Wu J
Wed May 01 00:00:00 UTC 2019
Orientin Improves Cognition by Enhancing Autophagosome Clearance in an Alzheimer's Mouse Model.
Journal of molecular neuroscience : MN
Zhong Y,Zheng QY,Sun CY,Zhang Z,Han K,Jia N
Tue Oct 01 00:00:00 UTC 2019
B-cell lymphoma-2-associated transcription factor 1 is overexpressed and contributes to sorafenib resistance in hepatocellular carcinoma.
Hepatology research : the official journal of the Japan Society of Hepatology
Yu S,Wang X,Dou N,Zhou J,Gao Y,Li Y
Fri Nov 01 00:00:00 UTC 2019
PGC-1α acts as an mediator of Sirtuin2 to protect annulus fibrosus from apoptosis induced by oxidative stress through restraining mitophagy.
International journal of biological macromolecules
Xu WN,Yang RZ,Zheng HL,Yu W,Zheng XF,Li B,Jiang SD,Jiang LS
Sun Sep 01 00:00:00 UTC 2019
PGC-1α acts as an mediator of Sirtuin2 to protect annulus fibrosus from apoptosis induced by oxidative stress through restraining mitophagy.
International journal of biological macromolecules
Xu WN,Yang RZ,Zheng HL,Yu W,Zheng XF,Li B,Jiang SD,Jiang LS
Sun Sep 01 00:00:00 UTC 2019
Molecular mechanism of miR-204 regulates proliferation, apoptosis and autophagy of cervical cancer cells by targeting ATF2.
Artificial cells, nanomedicine, and biotechnology
Li N,Guo X,Liu L,Wang L,Cheng R
Sun Dec 01 00:00:00 UTC 2019
Lyn Kinase Promotes the Proliferation of Malignant Melanoma Cells through Inhibition of Apoptosis and Autophagy via the PI3K/Akt Signaling Pathway.
Journal of Cancer
Zhang Q,Meng X,Qin G,Xue X,Dang N
Fri Oct 06 00:00:00 UTC 2023
LncRNA expression profile during autophagy and Malat1 function in macrophages.
PloS one
Ma Z,Zhang J,Xu X,Qu Y,Dong H,Dang J,Huo Z,Xu G
Thu Apr 02 00:00:00 UTC 2020
Diosbulbin B-Induced Mitochondria-Dependent Apoptosis in L-02 Hepatocytes is Regulated by Reactive Oxygen Species-Mediated Autophagy.
Frontiers in pharmacology
Ye J,Xue M,Liu Y,Zhu S,Li Y,Liu X,Cai D,Rui J,Zhang L
Wed Sep 30 00:00:00 UTC 2020
Bone marrow mesenchymal stem cells repair Cr (VI)- injured kidney by regulating mitochondria-mediated apoptosis and mitophagy mediated via the MAPK signaling pathway.
Ecotoxicology and environmental safety
Yin F,Yan J,Zhao Y,Guo KJ,Zhang ZL,Li AP,Meng CY,Guo L
Tue Jul 30 00:00:00 UTC 2019
Hexabromocyclododecanes promoted autophagy through the PI3K/Akt/mTOR pathway in L02 cells.
Journal of environmental management
Jin Y,Shang Y,Zhang D,An J,Pan D
Thu Aug 15 00:00:00 UTC 2019
Downregulation of microRNA-30a-5p contributes to the replication of duck enteritis virus by regulating Beclin-1-mediated autophagy.
Virology journal
Wu X,Jia R,Wang M,Chen S,Liu M,Zhu D,Zhao X,Yang Q,Wu Y,Yin Z,Zhang S,Huang J,Zhang L,Liu Y,Yu Y,Pan L,Tian B,Rehman MU,Chen X,Cheng A
Tue Nov 26 00:00:00 UTC 2019
Leucine-rich repeat and sterile alpha motif containing 1 promotes the oncogenic growth of human hepatocellular carcinoma cells.
Cancer cell international
Pian L,Huang X,Zhao M,Zhang Y,Qin C,Zhang J,Zhang J,Wang Q
Sun Apr 10 00:00:00 UTC 2022
miR-100-5p-abundant exosomes derived from infrapatellar fat pad MSCs protect articular cartilage and ameliorate gait abnormalities via inhibition of mTOR in osteoarthritis.
Biomaterials
Wu J,Kuang L,Chen C,Yang J,Zeng WN,Li T,Chen H,Huang S,Fu Z,Li J,Liu R,Ni Z,Chen L,Yang L
Sat Jun 01 00:00:00 UTC 2019
miR-100-5p-abundant exosomes derived from infrapatellar fat pad MSCs protect articular cartilage and ameliorate gait abnormalities via inhibition of mTOR in osteoarthritis.
Biomaterials
Wu J,Kuang L,Chen C,Yang J,Zeng WN,Li T,Chen H,Huang S,Fu Z,Li J,Liu R,Ni Z,Chen L,Yang L
Sat Jun 01 00:00:00 UTC 2019
Silver nanoparticles induce protective autophagy via Ca2+/CaMKKβ/AMPK/mTOR pathway in SH-SY5Y cells and rat brains.
Nanotoxicology
Li L,Li L,Zhou X,Yu Y,Li Z,Zuo D,Wu Y
Mon Apr 01 00:00:00 UTC 2019
Ferritinophagic Flux Activation in CT26 Cells Contributed to EMT Inhibition Induced by a Novel Iron Chelator, DpdtpA.
Oxidative medicine and cellular longevity
Sun Y,Li C,Feng J,Li Y,Zhai X,Zhang L,Li C
Fri Feb 21 00:00:00 UTC 2020
Rutin suppresses FNDC1 expression in bone marrow mesenchymal stem cells to inhibit postmenopausal osteoporosis.
American journal of translational research
Xiao Y,Wei R,Yuan Z,Lan X,Kuang J,Hu D,Song Y,Luo J
Thu Oct 01 00:00:00 UTC 2020
2,3',4,4',5-Pentachlorobiphenyl induced autophagy of the thyrocytes via DAPK2/PKD/VPS34 pathway.
Archives of toxicology
Zhou Q,Wang L,Chen H,Xu B,Xu W,Sheng Y,Duan Y
Sat Jun 01 00:00:00 UTC 2019
FePt/GO Nanosheets Suppress Proliferation, Enhance Radiosensitization and Induce Autophagy of Human Non-Small Cell Lung Cancer Cells.
International journal of biological sciences
Ma S,Miao H,Luo Y,Sun Y,Tian X,Wang F,You C,Peng S,Tang G,Yang C,Sun W,Li S,Mao Y,Xu J,Xiao Y,Gong Y,Quan H,Xie C
Mon Mar 16 00:00:00 UTC 2020
Alleviation of Hepatic Ischemia Reperfusion Injury by Oleanolic Acid Pretreating via Reducing HMGB1 Release and Inhibiting Apoptosis and Autophagy.
Mediators of inflammation
Wang W,Wu L,Li J,Ji J,Chen K,Yu Q,Li S,Feng J,Liu T,Zhang J,Chen J,Zhou Y,Mao Y,Wang F,Dai W,Fan X,Guo C,Wu J
Mon Feb 24 00:00:00 UTC 2020
Neuroprotective effect of Verbascoside on hypoxic-ischemic brain damage in neonatal rat.
Neuroscience letters
Wei W,Lu M,Lan X,Liu N,Wang H,Du J,Sun T,Li Y,Yu J
Tue Oct 15 00:00:00 UTC 2019
Neuroprotective effect of Verbascoside on hypoxic-ischemic brain damage in neonatal rat.
Neuroscience letters
Wei W,Lu M,Lan X,Liu N,Wang H,Du J,Sun T,Li Y,Yu J
Tue Oct 15 00:00:00 UTC 2019
Engagement of circular RNA HECW2 in the nonautophagic role of ATG5 implicated in the endothelial-mesenchymal transition.
Autophagy
Yang L,Han B,Zhang Y,Bai Y,Chao J,Hu G,Yao H
Fri Mar 22 00:00:00 UTC 2019
Engagement of circular RNA HECW2 in the nonautophagic role of ATG5 implicated in the endothelial-mesenchymal transition.
Autophagy
Yang L,Han B,Zhang Y,Bai Y,Chao J,Hu G,Yao H
Fri Mar 22 00:00:00 UTC 2019
Antagonizing peroxisome proliferator-activated receptor γ facilitates M1-to-M2 shift of microglia by enhancing autophagy via the LKB1-AMPK signaling pathway.
Aging cell
Ji J,Xue TF,Guo XD,Yang J,Guo RB,Wang J,Huang JY,Zhao XJ,Sun XL
Wed Aug 01 00:00:00 UTC 2018
Antagonizing peroxisome proliferator-activated receptor γ facilitates M1-to-M2 shift of microglia by enhancing autophagy via the LKB1-AMPK signaling pathway.
Aging cell
Ji J,Xue TF,Guo XD,Yang J,Guo RB,Wang J,Huang JY,Zhao XJ,Sun XL
Wed Aug 01 00:00:00 UTC 2018
Perigestational exposure to low doses of PBDE-47 induces excessive ER stress, defective autophagy and the resultant apoptosis contributing to maternal thyroid toxicity.
The Science of the total environment
Li P,Liu L,Zhou G,Tian Z,Luo C,Xia T,Chen J,Niu Q,Dong L,Zhao Q,Wang A,Zhang S
Sat Dec 15 00:00:00 UTC 2018
Perigestational exposure to low doses of PBDE-47 induces excessive ER stress, defective autophagy and the resultant apoptosis contributing to maternal thyroid toxicity.
The Science of the total environment
Li P,Liu L,Zhou G,Tian Z,Luo C,Xia T,Chen J,Niu Q,Dong L,Zhao Q,Wang A,Zhang S
Sat Dec 15 00:00:00 UTC 2018
Overcoming resistance to mitochondrial apoptosis by BZML-induced mitotic catastrophe is enhanced by inhibition of autophagy in A549/Taxol cells.
Cell proliferation
Bai Z,Gao M,Xu X,Zhang H,Xu J,Guan Q,Wang Q,Du J,Li Z,Zuo D,Zhang W,Wu Y
Wed Aug 01 00:00:00 UTC 2018
Overcoming resistance to mitochondrial apoptosis by BZML-induced mitotic catastrophe is enhanced by inhibition of autophagy in A549/Taxol cells.
Cell proliferation
Bai Z,Gao M,Xu X,Zhang H,Xu J,Guan Q,Wang Q,Du J,Li Z,Zuo D,Zhang W,Wu Y
Wed Aug 01 00:00:00 UTC 2018
Migration inhibition of water stress proteins from Nostoc commune Vauch. via activation of autophagy in DLD-1 cells.
International journal of biological macromolecules
Guo S,Liang X,Guo M,Zhang X,Li Z
Thu Nov 01 00:00:00 UTC 2018
Migration inhibition of water stress proteins from Nostoc commune Vauch. via activation of autophagy in DLD-1 cells.
International journal of biological macromolecules
Guo S,Liang X,Guo M,Zhang X,Li Z
Thu Nov 01 00:00:00 UTC 2018
Autophagy inhibition attenuates the induction of anti-inflammatory effect of catalpol in liver fibrosis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu Z,Zhu P,Zhang L,Xiong B,Tao J,Guan W,Li C,Chen C,Gu J,Duanmu J,Zhang W
Sun Jul 01 00:00:00 UTC 2018
Autophagy inhibition attenuates the induction of anti-inflammatory effect of catalpol in liver fibrosis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu Z,Zhu P,Zhang L,Xiong B,Tao J,Guan W,Li C,Chen C,Gu J,Duanmu J,Zhang W
Sun Jul 01 00:00:00 UTC 2018
Autophagy inhibition attenuates the induction of anti-inflammatory effect of catalpol in liver fibrosis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu Z,Zhu P,Zhang L,Xiong B,Tao J,Guan W,Li C,Chen C,Gu J,Duanmu J,Zhang W
Sun Jul 01 00:00:00 UTC 2018
Autophagy inhibition attenuates the induction of anti-inflammatory effect of catalpol in liver fibrosis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu Z,Zhu P,Zhang L,Xiong B,Tao J,Guan W,Li C,Chen C,Gu J,Duanmu J,Zhang W
Sun Jul 01 00:00:00 UTC 2018
Growth Inhibition of a Novel Iron Chelator, DpdtC, against Hepatoma Carcinoma Cell Lines Partly Attributed to Ferritinophagy-Mediated Lysosomal ROS Generation.
Oxidative medicine and cellular longevity
Huang T,Sun Y,Li Y,Wang T,Fu Y,Li C,Li C
Wed Dec 19 00:00:00 UTC 2018
Growth Inhibition of a Novel Iron Chelator, DpdtC, against Hepatoma Carcinoma Cell Lines Partly Attributed to Ferritinophagy-Mediated Lysosomal ROS Generation.
Oxidative medicine and cellular longevity
Huang T,Sun Y,Li Y,Wang T,Fu Y,Li C,Li C
Wed Dec 19 00:00:00 UTC 2018
Growth Inhibition of a Novel Iron Chelator, DpdtC, against Hepatoma Carcinoma Cell Lines Partly Attributed to Ferritinophagy-Mediated Lysosomal ROS Generation.
Oxidative medicine and cellular longevity
Huang T,Sun Y,Li Y,Wang T,Fu Y,Li C,Li C
Wed Dec 19 00:00:00 UTC 2018
Growth Inhibition of a Novel Iron Chelator, DpdtC, against Hepatoma Carcinoma Cell Lines Partly Attributed to Ferritinophagy-Mediated Lysosomal ROS Generation.
Oxidative medicine and cellular longevity
Huang T,Sun Y,Li Y,Wang T,Fu Y,Li C,Li C
Wed Dec 19 00:00:00 UTC 2018
Role of p62/SQSTM1 in lipopolysaccharide (LPS)-induced mucus hypersecretion in bronchial epithelial cells.
Life sciences
Wu Y,Li Y,Wang B,He X,Li Y,Wu B,Yu G,Wang H,Xu B
Mon Oct 15 00:00:00 UTC 2018
Role of p62/SQSTM1 in lipopolysaccharide (LPS)-induced mucus hypersecretion in bronchial epithelial cells.
Life sciences
Wu Y,Li Y,Wang B,He X,Li Y,Wu B,Yu G,Wang H,Xu B
Mon Oct 15 00:00:00 UTC 2018
Modulation of autophagy in the protective effect of resveratrol on PM2.5-induced pulmonary oxidative injury in mice.
Phytotherapy research : PTR
Li Y,Fu S,Li E,Sun X,Xu H,Meng Y,Wang X,Chen Y,Xie C,Geng S,Wu J,Zhong C,Xu P
Sat Dec 01 00:00:00 UTC 2018
Modulation of autophagy in the protective effect of resveratrol on PM2.5-induced pulmonary oxidative injury in mice.
Phytotherapy research : PTR
Li Y,Fu S,Li E,Sun X,Xu H,Meng Y,Wang X,Chen Y,Xie C,Geng S,Wu J,Zhong C,Xu P
Sat Dec 01 00:00:00 UTC 2018
Ginsenoside Rg5 induces apoptosis and autophagy via the inhibition of the PI3K/Akt pathway against breast cancer in a mouse model.
Food & function
Liu Y,Fan D
Wed Nov 14 00:00:00 UTC 2018
Ginsenoside Rg5 induces apoptosis and autophagy via the inhibition of the PI3K/Akt pathway against breast cancer in a mouse model.
Food & function
Liu Y,Fan D
Wed Nov 14 00:00:00 UTC 2018
Ginsenoside Rg5 induces apoptosis and autophagy via the inhibition of the PI3K/Akt pathway against breast cancer in a mouse model.
Food & function
Liu Y,Fan D
Wed Nov 14 00:00:00 UTC 2018
Ginsenoside Rg5 induces apoptosis and autophagy via the inhibition of the PI3K/Akt pathway against breast cancer in a mouse model.
Food & function
Liu Y,Fan D
Wed Nov 14 00:00:00 UTC 2018
Arsenic induces autophagy in developmental mouse cerebral cortex and hippocampus by inhibiting PI3K/Akt/mTOR signaling pathway: involvement of blood-brain barrier's tight junction proteins.
Archives of toxicology
Manthari RK,Tikka C,Ommati MM,Niu R,Sun Z,Wang J,Zhang J,Wang J
Thu Nov 01 00:00:00 UTC 2018
Arsenic induces autophagy in developmental mouse cerebral cortex and hippocampus by inhibiting PI3K/Akt/mTOR signaling pathway: involvement of blood-brain barrier's tight junction proteins.
Archives of toxicology
Manthari RK,Tikka C,Ommati MM,Niu R,Sun Z,Wang J,Zhang J,Wang J
Thu Nov 01 00:00:00 UTC 2018
Arsenic-Induced Autophagy in the Developing Mouse Cerebellum: Involvement of the Blood-Brain Barrier's Tight-Junction Proteins and the PI3K-Akt-mTOR Signaling Pathway.
Journal of agricultural and food chemistry
Manthari RK,Tikka C,Ommati MM,Niu R,Sun Z,Wang J,Zhang J,Wang J
Wed Aug 15 00:00:00 UTC 2018
Arsenic-Induced Autophagy in the Developing Mouse Cerebellum: Involvement of the Blood-Brain Barrier's Tight-Junction Proteins and the PI3K-Akt-mTOR Signaling Pathway.
Journal of agricultural and food chemistry
Manthari RK,Tikka C,Ommati MM,Niu R,Sun Z,Wang J,Zhang J,Wang J
Wed Aug 15 00:00:00 UTC 2018
Gli2 modulates cell cycle re-entry through autophagy-mediated regulation of the length of primary cilia.
Journal of cell science
Hsiao CJ,Chang CH,Ibrahim RB,Lin IH,Wang CH,Wang WJ,Tsai JW
Mon Dec 17 00:00:00 UTC 2018
Gli2 modulates cell cycle re-entry through autophagy-mediated regulation of the length of primary cilia.
Journal of cell science
Hsiao CJ,Chang CH,Ibrahim RB,Lin IH,Wang CH,Wang WJ,Tsai JW
Mon Dec 17 00:00:00 UTC 2018
The Role of SIRT1 in Autophagy in Lipopolysaccharide-Induced Mouse Type II Alveolar Epithelial Cells.
Inflammation
Liu J,Lv X,Dong W,Hu M,Xu J,Qian G,Li Y
Sat Dec 01 00:00:00 UTC 2018
The Role of SIRT1 in Autophagy in Lipopolysaccharide-Induced Mouse Type II Alveolar Epithelial Cells.
Inflammation
Liu J,Lv X,Dong W,Hu M,Xu J,Qian G,Li Y
Sat Dec 01 00:00:00 UTC 2018
Autophagy participates in innate immune defense in lamprey.
Fish & shellfish immunology
Luo L,Lu J,Wang Q,Chen S,Xu A,Yuan S
Sat Dec 01 00:00:00 UTC 2018
Autophagy participates in innate immune defense in lamprey.
Fish & shellfish immunology
Luo L,Lu J,Wang Q,Chen S,Xu A,Yuan S
Sat Dec 01 00:00:00 UTC 2018
The Protective Effects of Levo-Tetrahydropalmatine on ConA-Induced Liver Injury Are via TRAF6/JNK Signaling.
Mediators of inflammation
Yu Q,Liu T,Li S,Feng J,Wu L,Wang W,Chen K,Xia Y,Niu P,Xu L,Wang F,Dai W,Zhou Y,Guo C
Fri Apr 19 00:00:00 UTC 2019
The Protective Effects of Levo-Tetrahydropalmatine on ConA-Induced Liver Injury Are via TRAF6/JNK Signaling.
Mediators of inflammation
Yu Q,Liu T,Li S,Feng J,Wu L,Wang W,Chen K,Xia Y,Niu P,Xu L,Wang F,Dai W,Zhou Y,Guo C
Fri Apr 19 00:00:00 UTC 2019
The Hepatoprotection by Oleanolic Acid Preconditioning: Focusing on PPARα Activation.
PPAR research
Wang W,Chen K,Xia Y,Mo W,Wang F,Dai W,Niu P
Thu Mar 31 00:00:00 UTC 2022
The Hepatoprotection by Oleanolic Acid Preconditioning: Focusing on PPARα Activation.
PPAR research
Wang W,Chen K,Xia Y,Mo W,Wang F,Dai W,Niu P
Thu Mar 31 00:00:00 UTC 2022
The Hepatoprotection by Oleanolic Acid Preconditioning: Focusing on PPARα Activation.
PPAR research
Wang W,Chen K,Xia Y,Mo W,Wang F,Dai W,Niu P
Thu Mar 31 00:00:00 UTC 2022
The Hepatoprotection by Oleanolic Acid Preconditioning: Focusing on PPARα Activation.
PPAR research
Wang W,Chen K,Xia Y,Mo W,Wang F,Dai W,Niu P
Thu Mar 31 00:00:00 UTC 2022
Oxidative stress-mediated autophagic cell death participates in the neurotoxic effect on SH-SY5Y cells induced by excessive iodide.
Environmental toxicology
Liu H,Wang L,Zeng Q,Zhao L,Cui Y,Hou C,Zhang B,Zhang Z,Zhang S,Chen X,Wang A
Tue Jun 19 00:00:00 UTC 2018
Oxidative stress-mediated autophagic cell death participates in the neurotoxic effect on SH-SY5Y cells induced by excessive iodide.
Environmental toxicology
Liu H,Wang L,Zeng Q,Zhao L,Cui Y,Hou C,Zhang B,Zhang Z,Zhang S,Chen X,Wang A
Tue Jun 19 00:00:00 UTC 2018
Lycium barbarum polysaccharide attenuates diabetic testicular dysfunction via inhibition of the PI3K/Akt pathway-mediated abnormal autophagy in male mice.
Cell and tissue research
Shi GJ,Zheng J,Han XX,Jiang YP,Li ZM,Wu J,Chang Q,Niu Y,Sun T,Li YX,Chen Z,Yu JQ
Sat Dec 01 00:00:00 UTC 2018
Lycium barbarum polysaccharide attenuates diabetic testicular dysfunction via inhibition of the PI3K/Akt pathway-mediated abnormal autophagy in male mice.
Cell and tissue research
Shi GJ,Zheng J,Han XX,Jiang YP,Li ZM,Wu J,Chang Q,Niu Y,Sun T,Li YX,Chen Z,Yu JQ
Sat Dec 01 00:00:00 UTC 2018
Resveratrol relieves particulate matter (mean diameter < 2.5 μm)-induced oxidative injury of lung cells through attenuation of autophagy deregulation.
Journal of applied toxicology : JAT
Li Y,Qian W,Wang D,Meng Y,Wang X,Chen Y,Li X,Xie C,Zhong C,Fu S
Sat Sep 01 00:00:00 UTC 2018
Resveratrol relieves particulate matter (mean diameter < 2.5 μm)-induced oxidative injury of lung cells through attenuation of autophagy deregulation.
Journal of applied toxicology : JAT
Li Y,Qian W,Wang D,Meng Y,Wang X,Chen Y,Li X,Xie C,Zhong C,Fu S
Sat Sep 01 00:00:00 UTC 2018
Resveratrol relieves particulate matter (mean diameter < 2.5 μm)-induced oxidative injury of lung cells through attenuation of autophagy deregulation.
Journal of applied toxicology : JAT
Li Y,Qian W,Wang D,Meng Y,Wang X,Chen Y,Li X,Xie C,Zhong C,Fu S
Sat Sep 01 00:00:00 UTC 2018
Resveratrol relieves particulate matter (mean diameter < 2.5 μm)-induced oxidative injury of lung cells through attenuation of autophagy deregulation.
Journal of applied toxicology : JAT
Li Y,Qian W,Wang D,Meng Y,Wang X,Chen Y,Li X,Xie C,Zhong C,Fu S
Sat Sep 01 00:00:00 UTC 2018
Protective effects of α‑2‑macroglobulin on human bone marrow mesenchymal stem cells in radiation injury.
Molecular medicine reports
Liu Y,Cao W,Kong X,Li J,Chen X,Ge Y,Zhong W,Fang S
Thu Nov 01 00:00:00 UTC 2018
Protective effects of α‑2‑macroglobulin on human bone marrow mesenchymal stem cells in radiation injury.
Molecular medicine reports
Liu Y,Cao W,Kong X,Li J,Chen X,Ge Y,Zhong W,Fang S
Thu Nov 01 00:00:00 UTC 2018
Rapamycin and FTY720 Alleviate Atherosclerosis by Cross Talk of Macrophage Polarization and Autophagy.
BioMed research international
Sun RZ,Fan Y,Liang X,Gong TT,Wang Q,Liu H,Shan ZY,Lei L
Wed Apr 24 00:00:00 UTC 2019
Rapamycin and FTY720 Alleviate Atherosclerosis by Cross Talk of Macrophage Polarization and Autophagy.
BioMed research international
Sun RZ,Fan Y,Liang X,Gong TT,Wang Q,Liu H,Shan ZY,Lei L
Wed Apr 24 00:00:00 UTC 2019
Cantharidin suppresses cell growth and migration, and activates autophagy in human non-small cell lung cancer cells.
Oncology letters
Liu YP,Li L,Xu L,Dai EN,Chen WD
Tue May 01 00:00:00 UTC 2018
Cantharidin suppresses cell growth and migration, and activates autophagy in human non-small cell lung cancer cells.
Oncology letters
Liu YP,Li L,Xu L,Dai EN,Chen WD
Tue May 01 00:00:00 UTC 2018
Salidroside mediates apoptosis and autophagy inhibition in concanavalin A-induced liver injury.
Experimental and therapeutic medicine
Feng J,Niu P,Chen K,Wu L,Liu T,Xu S,Li J,Li S,Wang W,Lu X,Yu Q,Liu N,Xu L,Wang F,Dai W,Xia Y,Fan X,Guo C
Fri Jun 01 00:00:00 UTC 2018
Salidroside mediates apoptosis and autophagy inhibition in concanavalin A-induced liver injury.
Experimental and therapeutic medicine
Feng J,Niu P,Chen K,Wu L,Liu T,Xu S,Li J,Li S,Wang W,Lu X,Yu Q,Liu N,Xu L,Wang F,Dai W,Xia Y,Fan X,Guo C
Fri Jun 01 00:00:00 UTC 2018
Salidroside mediates apoptosis and autophagy inhibition in concanavalin A-induced liver injury.
Experimental and therapeutic medicine
Feng J,Niu P,Chen K,Wu L,Liu T,Xu S,Li J,Li S,Wang W,Lu X,Yu Q,Liu N,Xu L,Wang F,Dai W,Xia Y,Fan X,Guo C
Fri Jun 01 00:00:00 UTC 2018
Salidroside mediates apoptosis and autophagy inhibition in concanavalin A-induced liver injury.
Experimental and therapeutic medicine
Feng J,Niu P,Chen K,Wu L,Liu T,Xu S,Li J,Li S,Wang W,Lu X,Yu Q,Liu N,Xu L,Wang F,Dai W,Xia Y,Fan X,Guo C
Fri Jun 01 00:00:00 UTC 2018
Xiaoqinglong Decoction Attenuates Chronic Obstructive Pulmonary Disease in Rats via Inhibition of Autophagy.
Evidence-based complementary and alternative medicine : eCAM
Wang H,Mao B,Chen C
Tue Sep 29 00:00:00 UTC 2020
Xiaoqinglong Decoction Attenuates Chronic Obstructive Pulmonary Disease in Rats via Inhibition of Autophagy.
Evidence-based complementary and alternative medicine : eCAM
Wang H,Mao B,Chen C
Tue Sep 29 00:00:00 UTC 2020
Mutant UBQLN2P497H in motor neurons leads to ALS-like phenotypes and defective autophagy in rats.
Acta neuropathologica communications
Chen T,Huang B,Shi X,Gao L,Huang C
Thu Nov 08 00:00:00 UTC 2018
Mutant UBQLN2P497H in motor neurons leads to ALS-like phenotypes and defective autophagy in rats.
Acta neuropathologica communications
Chen T,Huang B,Shi X,Gao L,Huang C
Thu Nov 08 00:00:00 UTC 2018
Triptolide protects podocytes via autophagy in immunoglobulin A nephropathy.
Experimental and therapeutic medicine
Liang S,Jin J,Shen X,Jiang X,Li Y,He Q
Sat Sep 01 00:00:00 UTC 2018
Rapamycin Confers Neuroprotection against Colistin-Induced Oxidative Stress, Mitochondria Dysfunction, and Apoptosis through the Activation of Autophagy and mTOR/Akt/CREB Signaling Pathways.
ACS chemical neuroscience
Dai C,Ciccotosto GD,Cappai R,Wang Y,Tang S,Hoyer D,Schneider EK,Velkov T,Xiao X
Wed Apr 18 00:00:00 UTC 2018
Rapamycin Confers Neuroprotection against Colistin-Induced Oxidative Stress, Mitochondria Dysfunction, and Apoptosis through the Activation of Autophagy and mTOR/Akt/CREB Signaling Pathways.
ACS chemical neuroscience
Dai C,Ciccotosto GD,Cappai R,Wang Y,Tang S,Hoyer D,Schneider EK,Velkov T,Xiao X
Wed Apr 18 00:00:00 UTC 2018
Development of the Swimbladder Surfactant System and Biogenesis of Lysosome-Related Organelles Is Regulated by BLOS1 in Zebrafish.
Genetics
Chen T,Song G,Yang H,Mao L,Cui Z,Huang K
Thu Mar 01 00:00:00 UTC 2018
Autophagy is positively associated with the accumulation of myeloid‑derived suppressor cells in 4‑nitroquinoline‑1‑oxide‑induced oral cancer.
Oncology reports
Wu JS,Li L,Wang SS,Pang X,Wu JB,Sheng SR,Tang YJ,Tang YL,Zheng M,Liang XH
Sat Dec 01 00:00:00 UTC 2018
Autophagy is positively associated with the accumulation of myeloid‑derived suppressor cells in 4‑nitroquinoline‑1‑oxide‑induced oral cancer.
Oncology reports
Wu JS,Li L,Wang SS,Pang X,Wu JB,Sheng SR,Tang YJ,Tang YL,Zheng M,Liang XH
Sat Dec 01 00:00:00 UTC 2018
Autophagy is positively associated with the accumulation of myeloid‑derived suppressor cells in 4‑nitroquinoline‑1‑oxide‑induced oral cancer.
Oncology reports
Wu JS,Li L,Wang SS,Pang X,Wu JB,Sheng SR,Tang YJ,Tang YL,Zheng M,Liang XH
Sat Dec 01 00:00:00 UTC 2018
Interplay of Autophagy Inducer Rapamycin and Proteasome Inhibitor MG132 in Reduction of Foam Cell Formation and Inflammatory Cytokine Expression.
Cell transplantation
Zhang W,Xu W,Chen W,Zhou Q
Wed Aug 01 00:00:00 UTC 2018
Autophagy defects and related genetic variations in renal cell carcinoma with eosinophilic cytoplasmic inclusions.
Scientific reports
Yu Z,Ma J,Li X,Liu Y,Li M,Wang L,Zhao M,He H,Zhang Y,Rao Q,Zhao D,Wang Y,Fan L,Li P,Liu Y,Liu F,Zhang F,Ye J,Yan Q,Guo S,Wang Z
Mon Jul 02 00:00:00 UTC 2018
Autophagy defects and related genetic variations in renal cell carcinoma with eosinophilic cytoplasmic inclusions.
Scientific reports
Yu Z,Ma J,Li X,Liu Y,Li M,Wang L,Zhao M,He H,Zhang Y,Rao Q,Zhao D,Wang Y,Fan L,Li P,Liu Y,Liu F,Zhang F,Ye J,Yan Q,Guo S,Wang Z
Mon Jul 02 00:00:00 UTC 2018
Autophagy defects and related genetic variations in renal cell carcinoma with eosinophilic cytoplasmic inclusions.
Scientific reports
Yu Z,Ma J,Li X,Liu Y,Li M,Wang L,Zhao M,He H,Zhang Y,Rao Q,Zhao D,Wang Y,Fan L,Li P,Liu Y,Liu F,Zhang F,Ye J,Yan Q,Guo S,Wang Z
Mon Jul 02 00:00:00 UTC 2018
Salidroside ameliorates autophagy and activation of hepatic stellate cells in mice via NF-κB and TGF-β1/Smad3 pathways.
Drug design, development and therapy
Feng J,Chen K,Xia Y,Wu L,Li J,Li S,Wang W,Lu X,Liu T,Guo C
Wed Oct 31 00:00:00 UTC 2018
Salidroside ameliorates autophagy and activation of hepatic stellate cells in mice via NF-κB and TGF-β1/Smad3 pathways.
Drug design, development and therapy
Feng J,Chen K,Xia Y,Wu L,Li J,Li S,Wang W,Lu X,Liu T,Guo C
Wed Oct 31 00:00:00 UTC 2018
Inhibition of insulin resistance by PGE1 via autophagy-dependent FGF21 pathway in diabetic nephropathy.
Scientific reports
Wei W,An XR,Jin SJ,Li XX,Xu M
Mon Jan 08 00:00:00 UTC 2018
Inhibition of insulin resistance by PGE1 via autophagy-dependent FGF21 pathway in diabetic nephropathy.
Scientific reports
Wei W,An XR,Jin SJ,Li XX,Xu M
Mon Jan 08 00:00:00 UTC 2018
Peroxisome Proliferator-Activated Receptor (PPARγ) Plays a Protective Role in Cigarette Smoking-Induced Inflammation via AMP-Activated Protein Kinase (AMPK) Signaling.
Medical science monitor : international medical journal of experimental and clinical research
Wang L,Yin Y,Hou G,Kang J,Wang Q
Wed Jul 25 00:00:00 UTC 2018
Peroxisome Proliferator-Activated Receptor (PPARγ) Plays a Protective Role in Cigarette Smoking-Induced Inflammation via AMP-Activated Protein Kinase (AMPK) Signaling.
Medical science monitor : international medical journal of experimental and clinical research
Wang L,Yin Y,Hou G,Kang J,Wang Q
Wed Jul 25 00:00:00 UTC 2018
Autophagy is required for proper meiosis of porcine oocytes maturing in vitro.
Scientific reports
Shen XH,Jin YX,Liang S,Kwon JW,Zhu JW,Lei L,Kim NH
Wed Aug 22 00:00:00 UTC 2018
Autophagy is required for proper meiosis of porcine oocytes maturing in vitro.
Scientific reports
Shen XH,Jin YX,Liang S,Kwon JW,Zhu JW,Lei L,Kim NH
Wed Aug 22 00:00:00 UTC 2018
β-Arrestin2 Inhibits Expression of Inflammatory Cytokines in BEAS-2B Lung Epithelial Cells Treated with Cigarette Smoke Condensate via Inhibition of Autophagy.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wu Y,Li Y,Wu B,Tan C,He X,Xu B,Yu G,Wang H
Fri Nov 09 00:00:00 UTC 2018
β-Arrestin2 Inhibits Expression of Inflammatory Cytokines in BEAS-2B Lung Epithelial Cells Treated with Cigarette Smoke Condensate via Inhibition of Autophagy.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wu Y,Li Y,Wu B,Tan C,He X,Xu B,Yu G,Wang H
Fri Nov 09 00:00:00 UTC 2018
Fluoride induces apoptosis and autophagy through the IL-17 signaling pathway in mice hepatocytes.
Archives of toxicology
Zhao Y,Li Y,Wang J,Manthari RK,Wang J
Thu Nov 01 00:00:00 UTC 2018
Isoliquiritigenin inhibits proliferation and metastasis of MKN28 gastric cancer cells by suppressing the PI3K/AKT/mTOR signaling pathway.
Molecular medicine reports
Zhang XR,Wang SY,Sun W,Wei C
Sat Sep 01 00:00:00 UTC 2018
Circular RNA HIPK2 regulates astrocyte activation via cooperation of autophagy and ER stress by targeting MIR124-2HG.
Autophagy
Huang R,Zhang Y,Han B,Bai Y,Zhou R,Gan G,Chao J,Hu G,Yao H
Tue Oct 03 00:00:00 UTC 2017
Circular RNA HIPK2 regulates astrocyte activation via cooperation of autophagy and ER stress by targeting MIR124-2HG.
Autophagy
Huang R,Zhang Y,Han B,Bai Y,Zhou R,Gan G,Chao J,Hu G,Yao H
Tue Oct 03 00:00:00 UTC 2017
Exercise Combined with Rhodiola sacra Supplementation Improves Exercise Capacity and Ameliorates Exhaustive Exercise-Induced Muscle Damage through Enhancement of Mitochondrial Quality Control.
Oxidative medicine and cellular longevity
Dun Y,Liu S,Zhang W,Xie M,Qiu L
Fri Aug 17 00:00:00 UTC 2018
Exercise Combined with Rhodiola sacra Supplementation Improves Exercise Capacity and Ameliorates Exhaustive Exercise-Induced Muscle Damage through Enhancement of Mitochondrial Quality Control.
Oxidative medicine and cellular longevity
Dun Y,Liu S,Zhang W,Xie M,Qiu L
Fri Aug 17 00:00:00 UTC 2018
Rosiglitazone Exerts an Anti-depressive Effect in Unpredictable Chronic Mild-Stress-Induced Depressive Mice by Maintaining Essential Neuron Autophagy and Inhibiting Excessive Astrocytic Apoptosis.
Frontiers in molecular neuroscience
Zhao Z,Zhang L,Guo XD,Cao LL,Xue TF,Zhao XJ,Yang DD,Yang J,Ji J,Huang JY,Sun XL
Wed Sep 30 00:00:00 UTC 2020
Rosiglitazone Exerts an Anti-depressive Effect in Unpredictable Chronic Mild-Stress-Induced Depressive Mice by Maintaining Essential Neuron Autophagy and Inhibiting Excessive Astrocytic Apoptosis.
Frontiers in molecular neuroscience
Zhao Z,Zhang L,Guo XD,Cao LL,Xue TF,Zhao XJ,Yang DD,Yang J,Ji J,Huang JY,Sun XL
Wed Sep 30 00:00:00 UTC 2020
Fluoride-Induced Autophagy via the Regulation of Phosphorylation of Mammalian Targets of Rapamycin in Mice Leydig Cells.
Journal of agricultural and food chemistry
Zhang J,Zhu Y,Shi Y,Han Y,Liang C,Feng Z,Zheng H,Eng M,Wang J
Wed Oct 11 00:00:00 UTC 2017
Fluoride-Induced Autophagy via the Regulation of Phosphorylation of Mammalian Targets of Rapamycin in Mice Leydig Cells.
Journal of agricultural and food chemistry
Zhang J,Zhu Y,Shi Y,Han Y,Liang C,Feng Z,Zheng H,Eng M,Wang J
Wed Oct 11 00:00:00 UTC 2017
Relationship between autophagy and perineural invasion, clinicopathological features, and prognosis in pancreatic cancer.
World journal of gastroenterology
Yang YH,Liu JB,Gui Y,Lei LL,Zhang SJ
Sat Oct 28 00:00:00 UTC 2017
Relationship between autophagy and perineural invasion, clinicopathological features, and prognosis in pancreatic cancer.
World journal of gastroenterology
Yang YH,Liu JB,Gui Y,Lei LL,Zhang SJ
Sat Oct 28 00:00:00 UTC 2017
Autophagy induction in the skeletal myogenic differentiation of human tonsil-derived mesenchymal stem cells.
International journal of molecular medicine
Park S,Choi Y,Jung N,Kim J,Oh S,Yu Y,Ahn JH,Jo I,Choi BO,Jung SC
Sat Apr 01 00:00:00 UTC 2017
Autophagy induction in the skeletal myogenic differentiation of human tonsil-derived mesenchymal stem cells.
International journal of molecular medicine
Park S,Choi Y,Jung N,Kim J,Oh S,Yu Y,Ahn JH,Jo I,Choi BO,Jung SC
Sat Apr 01 00:00:00 UTC 2017
Rab11a is required for porcine reproductive and respiratory syndrome virus induced autophagy to promote viral replication.
Biochemical and biophysical research communications
Wang K,Li S,Worku T,Hao X,Yang L,Zhang S
Sat Oct 14 00:00:00 UTC 2017
Rab11a is required for porcine reproductive and respiratory syndrome virus induced autophagy to promote viral replication.
Biochemical and biophysical research communications
Wang K,Li S,Worku T,Hao X,Yang L,Zhang S
Sat Oct 14 00:00:00 UTC 2017
Rab11a is required for porcine reproductive and respiratory syndrome virus induced autophagy to promote viral replication.
Biochemical and biophysical research communications
Wang K,Li S,Worku T,Hao X,Yang L,Zhang S
Sat Oct 14 00:00:00 UTC 2017
Rab11a is required for porcine reproductive and respiratory syndrome virus induced autophagy to promote viral replication.
Biochemical and biophysical research communications
Wang K,Li S,Worku T,Hao X,Yang L,Zhang S
Sat Oct 14 00:00:00 UTC 2017
Levels of Leydig cell autophagy regulate the fertility of male naked mole-rats.
Oncotarget
Yang W,Li L,Huang X,Kan G,Lin L,Cheng J,Xu C,Sun W,Cong W,Zhao S,Cui S
Fri Nov 17 00:00:00 UTC 2017
Levels of Leydig cell autophagy regulate the fertility of male naked mole-rats.
Oncotarget
Yang W,Li L,Huang X,Kan G,Lin L,Cheng J,Xu C,Sun W,Cong W,Zhao S,Cui S
Fri Nov 17 00:00:00 UTC 2017
Levels of Leydig cell autophagy regulate the fertility of male naked mole-rats.
Oncotarget
Yang W,Li L,Huang X,Kan G,Lin L,Cheng J,Xu C,Sun W,Cong W,Zhao S,Cui S
Fri Nov 17 00:00:00 UTC 2017
Levels of Leydig cell autophagy regulate the fertility of male naked mole-rats.
Oncotarget
Yang W,Li L,Huang X,Kan G,Lin L,Cheng J,Xu C,Sun W,Cong W,Zhao S,Cui S
Fri Nov 17 00:00:00 UTC 2017
Resveratrol prevents the combined maternal plus postweaning high-fat-diets-induced hypertension in male offspring.
The Journal of nutritional biochemistry
Tain YL,Lin YJ,Sheen JM,Lin IC,Yu HR,Huang LT,Hsu CN
Sun Oct 01 00:00:00 UTC 2017
Decreased Level of Klotho Contributes to Drug Resistance in Lung Cancer Cells: Involving in Klotho-Mediated Cell Autophagy.
DNA and cell biology
Chen T,Ren H,Thakur A,Yang T,Li Y,Zhang S,Wang T,Chen M
Thu Dec 01 00:00:00 UTC 2016
Decreased Level of Klotho Contributes to Drug Resistance in Lung Cancer Cells: Involving in Klotho-Mediated Cell Autophagy.
DNA and cell biology
Chen T,Ren H,Thakur A,Yang T,Li Y,Zhang S,Wang T,Chen M
Thu Dec 01 00:00:00 UTC 2016
Effects of the total saponins from Rosa laevigata Michx fruit against acetaminophen-induced liver damage in mice via induction of autophagy and suppression of inflammation and apoptosis.
Molecules (Basel, Switzerland)
Dong D,Xu L,Han X,Qi Y,Xu Y,Yin L,Liu K,Peng J
Fri May 30 00:00:00 UTC 2014
Cathepsin L stimulates autophagy and inhibits apoptosis of ox-LDL-induced endothelial cells: potential role in atherosclerosis.
International journal of molecular medicine
Wei DH,Jia XY,Liu YH,Guo FX,Tang ZH,Li XH,Wang Z,Liu LS,Wang GX,Jian ZS,Ruan CG
Fri Feb 01 00:00:00 UTC 2013
Cathepsin L stimulates autophagy and inhibits apoptosis of ox-LDL-induced endothelial cells: potential role in atherosclerosis.
International journal of molecular medicine
Wei DH,Jia XY,Liu YH,Guo FX,Tang ZH,Li XH,Wang Z,Liu LS,Wang GX,Jian ZS,Ruan CG
Fri Feb 01 00:00:00 UTC 2013
PM2.5-induced oxidative stress triggers autophagy in human lung epithelial A549 cells.
Toxicology in vitro : an international journal published in association with BIBRA
Deng X,Zhang F,Rui W,Long F,Wang L,Feng Z,Chen D,Ding W
Sun Sep 01 00:00:00 UTC 2013
PM2.5-induced oxidative stress triggers autophagy in human lung epithelial A549 cells.
Toxicology in vitro : an international journal published in association with BIBRA
Deng X,Zhang F,Rui W,Long F,Wang L,Feng Z,Chen D,Ding W
Sun Sep 01 00:00:00 UTC 2013
PM2.5-induced oxidative stress triggers autophagy in human lung epithelial A549 cells.
Toxicology in vitro : an international journal published in association with BIBRA
Deng X,Zhang F,Rui W,Long F,Wang L,Feng Z,Chen D,Ding W
Sun Sep 01 00:00:00 UTC 2013
PM2.5-induced oxidative stress triggers autophagy in human lung epithelial A549 cells.
Toxicology in vitro : an international journal published in association with BIBRA
Deng X,Zhang F,Rui W,Long F,Wang L,Feng Z,Chen D,Ding W
Sun Sep 01 00:00:00 UTC 2013