Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • LC3B Antibodies

            Invitrogen

            LC3B Polyclonal Antibody

            Advanced Verification
            2 Published Figures
            7 References
            View all (56) LC3B antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-32254
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            586.00
            100 µL
             
            In stock
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite LC3B Polyclonal Antibody

            • Antibody Testing Data (16)
            • Published Figures (2)
            • Advanced Verification (1)
            Application
            LC3B Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 19

            LC3B Antibody (PA5-32254) in ICC/IF

            Immunocytochemistry-Immunofluorescence analysis of LC3B in HeLa cells mock (left) and in HeLa cells treated with 50 µM Chloroquine for 24 hr (right) fixed with 4% paraformaldehyde at RT for 15 min. Green: LC3B Polyclonal Antibody (Product # PA5-32254) diluted at 1:2,000. Red: alpha Tubulin, a cytoskeleton marker, stained by alpha Tubulin antibody. Blue: Hoechst 33342 staining.
            View Product

            LC3B Antibody in Immunocytochemistry (ICC/IF)
            LC3B Antibody in Immunocytochemistry (ICC/IF)
            LC3B Antibody in Immunocytochemistry (ICC/IF)
            LC3B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Flow Cytometry (Flow)
            LC3B Antibody in Immunoprecipitation (IP)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody in Western Blot (WB)
            LC3B Antibody

            Product Details

            PA5-32254

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:3,000
            View 5 publications 5 publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100-1:1,000
            -

            Immunohistochemistry (Frozen) (IHC (F))

            Assay-dependent
            -

            Immunocytochemistry (ICC/IF)

            1:100-1:1,000
            View 1 publication 1 publication

            Flow Cytometry (Flow)

            1:50-1:200
            -

            Immunoprecipitation (IP)

            1:100-1:500
            -

            Immunomicroscopy (IM)

            Assay-dependent
            -
            Product Specifications

            Species Reactivity

            Human, Insect, Mouse, Pig, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human LC3B. The exact sequence is proprietary.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.34 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7, with 1% BSA, 20% glycerol

            Contains

            0.025% ProClin 300

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2549727

            Product Specific Information

            Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH, mouse brain, Rat brain, HepG2 (untreated), HepG2 (3 µM Thapsigargin treatment for 12 hr), HepG2 (3 µM Thapsigargin treatment for 16hr), HepG2 (3 µM Thapsigargin treatment for 24hr), Huh7 (un-infected), Huh7 (HCV-infected), HeLa, HeLa (50 µM Chloroquine treatment for 24 hr).

            Predicted reactivity: Zebrafish (100%), Japanese Medaka (100%), Pig (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%).

            Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

            Target Information

            LC3B (Autophagy Marker Light Chain 3B, MAP1A/MAP1B LC3 B) in humans, is encoded by the gene MAP1LC3B (Microtubule-associated proteins 1A/1B light chain 3B). LC3B is associated with microtubule assembly and important is in neurogenesis. Recent studies indicate that LC3B plays a vital role in autophagy, a process that involves the bulk degradation of cytoplasmic component. Three human LC3 isoforms undergo post-translational modifications during autophagy. Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). LC3B is a microtubule-associated protein that mediate the physical interactions between microtubules and components of the cytoskeleton. LC3B may play a role in processes involving cancer, aging, metabolic and neurodegenerative disorders and cardiovascular/pulmonary diseases.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Western Blot
            • Secondary Antibodies
            • Isotype Controls
            • Loading Controls WB
            • Electrophoresis WB
            • Transfer Stacks WB
            • Substrates WB
            • Labeling Kits WB
            Catalog #
            656140
            Invitrogen
            Goat anti-Rabbit IgG (H+L) Secondary Antibody, Biotin
            Unit Size :
            2 mL
            Catalog #
            31210
            Invitrogen
            Goat anti-Rabbit IgG (H+L) Secondary Antibody
            Unit Size :
            2 mg
            Catalog #
            A21441
            Invitrogen
            Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
            Unit Size :
            1 mg
            Catalog #
            A32794
            Invitrogen
            Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 555
            Unit Size :
            1 mg
            Catalog #
            A32732
            Invitrogen
            Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 555
            Unit Size :
            1 mg
            Catalog #
            A32735
            Invitrogen
            Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 800
            Unit Size :
            1 mg
            Catalog #
            31460
            Invitrogen
            Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP
            Unit Size :
            2 mL
            Catalog #
            A11012
            Invitrogen
            Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594
            Unit Size :
            1 mg
            Catalog #
            35552
            Invitrogen
            Goat anti-Rabbit IgG (H+L) Secondary Antibody, DyLight™ 488
            Unit Size :
            1 mL
            Catalog #
            A10042
            Invitrogen
            Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 568
            Unit Size :
            1 mg

            Bioinformatics

            Protein Aliases: ATG8E; Autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 B; LC3 beta; LC3A; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 B; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; MAP1BLC3; MAP1LC3A; Microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3 beta 2; microtubule-associated proteins 1A/1B light chain 3; Microtubule-associated proteins 1A/1B light chain 3 beta 2; Microtubule-associated proteins 1A/1B light chain 3B; Microtubule-associated proteins 1A/1B light chain 3B-like; Microtubule-associated proteins 1A/1B light chain 3B-like protein; RP11-346K17.1

            View more View less

            Gene Aliases: 1010001C15Rik; Atg8; ATG8F; ATG8G; LC3B; MAP1A/1BLC3; MAP1ALC3; Map1lc3; MAP1LC3B; MAP1LC3B-a; MAP1LC3B2; Mpl3; zbs559

            View more View less

            UniProt ID: (Human) Q9GZQ8, (Human) A6NCE7, (Rat) Q62625, (Mouse) Q9CQV6

            View more View less

            Entrez Gene ID: (Human) 81631, (Pig) 100462754, (Human) 643246, (Rat) 64862, (Mouse) 67443

            View more View less

            Function(s)

            protein binding microtubule binding ubiquitin protein ligase binding tubulin binding protein domain specific binding GABA receptor binding non-motor microtubule binding protein

            Process(es)

            autophagosome assembly cellular response to nitrogen starvation macroautophagy regulation of mitophagy mitophagy autophagy positive regulation of protein binding nucleophagy membrane fusion positive regulation of mucus secretion
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-584499df9d-pwm6p:80/100.66.79.199:80.
            git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.33.0-2025.07.30-1.0