Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGPPGSPWQW VTLLLGLLLP PAAPFWLLNV LFPPHTTPKA ELSNHTRPVI LVPGCLGNQL EAKLDKPDVV NWMCYRKTED FFTIWLDLNM FLPLGVDCWI DNTRVVYNRS SGLVSNAPGV QIRVPGFGKT YSVEYLDSSK LAGYLHTLVQ NLVNNGYVRD ETVRAAPYDW RLEPGQQEEY YRKLAGLVEE MHAAYGKPVF LIGHSLGCLH LLYFLLRQPQ AWKDRFIDGF ISLGAPWGGS IKPMLVLASG DNQGIPIMSS IKLKEEQRIT TTSPWMFPSR MAWPEDHVFI STPSFNYTGR DFQRFFADLH FEEGWYMWLQ SRDLLAGLPA PGVEVYCLYG VGLPTPRTYI YDHGFPYTDP VGVLYEDGDD TVATRSTELC GLWQGRQPQP VHLLPLHGIQ HLNMVFSNLT LEHINAILLG AYRQGPPASP TASPEPPPPE
Lecithin: cholesterol acyltransferase (LCAT) is the plasma enzyme responsible for most HDL cholesteryl esters in human plasma. In species with cholesteryl ester transfer protein (CETP) it is also responsible for a significant proportion of VLDL and LDL cholesteryl esters. LCAT, through the esterification of cholesterol, plays a role in the reverse cholesterol transport pathway by facilitating the net movement of cholesterol from peripheral tissues back to the liver for excretion. LCAT deficiency results in low levels of plasma HDL, whereas a transgenic overexpression of LCAT results in increased plasma HDL concentrations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-alkyl-2-acetylglycerophosphocholine esterase; Lecithin-cholesterol acyltransferase; PAF acetylhydrolase; phosphatidylcholine--sterol O-acyltransferase; Phosphatidylcholine-sterol acyltransferase; Phospholipid-cholesterol acyltransferase; Platelet-activating factor acetylhydrolase; testicular secretory protein Li 24
Gene Aliases: LCAT
UniProt ID: (Human) P04180
Entrez Gene ID: (Human) 3931
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support