Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: NT2D1, RD, mouse muscle, rat muscle.
Predicted reactivity: Mouse (97%), Rat (97%), Chicken (84%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a PDZ domain-containing protein. PDZ motifs are modular protein-protein interaction domains consisting of 80-120 amino acid residues. PDZ domain-containing proteins interact with each other in cytoskeletal assembly or with other proteins involved in targeting and clustering of membrane proteins. The protein encoded by this gene interacts with alpha-actinin-2 through its N-terminal PDZ domain and with protein kinase C via its C-terminal LIM domains. The LIM domain is a cysteine-rich motif defined by 50-60 amino acids containing two zinc-binding modules. This protein also interacts with all three members of the myozenin family. Mutations in this gene have been associated with myofibrillar myopathy and dilated cardiomyopathy. Alternatively spliced transcript variants encoding different isoforms have been identified; all isoforms have N-terminal PDZ domains while only longer isoforms (1, 2 and 5) have C-terminal LIM domains.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiomyopathy, dilated 1C (autosomal dominant); cypher; FLJ35865; HGNC:15710; KIAA01613; LDB3; LIM domain-binding protein 3; oracle protein; PDZ and LIM domain 6; PDZ-LIM domain protein; Protein cypher; Protein oracle; Z-band alternatively spliced PDZ motif protein; Z-band alternatively spliced PDZ-motif protein
Gene Aliases: AW742271; CMD1C; CMH24; CMPD3; CYPHER; KIAA0613; LDB3; LDB3Z1; LDB3Z4; LVNC3; MFM4; ORACLE; PDLIM6; RGD1564875; ZASP
UniProt ID: (Human) O75112, (Mouse) Q9R0Z1
Entrez Gene ID: (Human) 11155, (Mouse) 24131, (Rat) 498587
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support