Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MARLLRSATW ELFPWRGYCS QKAKGELCRD FVEALKAVVG GSHVSTAAVV REQHGRDESV HRCEPPDAVV WPQNVEQVSR LAALCYRQGV PIIPFGTGTG LEGGVCAVQG GVCVNLTHMD RILELNQEDF SVVVEPGVTR KALNAHLRDS GLWFPVDPGA DASLCGMAAT GASGTNAVRY GTMRDNVLNL EVVLPDGRLL HTAGRGRHFR FGFWPEIPHH TAWYSPCVSL GRRKSAAGYN LTGLFVGSEG TLGLITATTL RLHPAPEATV AATCAFPSVQ AAVDSTVHIL QAAVPVARIE FLDEVMMDAC NRYSKLNCLV APTLFLEFHG SQQALEEQLQ RTEEIVQQNG ASDFSWAKEA EERSRLWTAR HNAWYAALAT RPGCKGYSTD VCVPISRLPE IVVQTKEDLN ASGLTGSIVG HVGDGNFHCI LLVNPDDAEE LGRVKAFAEQ LGRRALALHG TCTGEHGIGM GKRQLLQEEV GAVGVETMRQ LKAVLDPQGL MNPGKVL
LDHD belongs to the D-isomer specific 2-hydroxyacid dehydrogenase family. The similar protein in yeast has both D-lactate and D-glycerate dehydrogenase activities. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-lactate dehydrogenase; DLD; LDHD; Probable D-lactate dehydrogenase, mitochondrial
Gene Aliases: DLD; LDHD
UniProt ID: (Human) Q86WU2
Entrez Gene ID: (Human) 197257
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support