Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
LDHD belongs to the D-isomer specific 2-hydroxyacid dehydrogenase family. The similar protein in yeast has both D-lactate and D-glycerate dehydrogenase activities. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-lactate dehydrogenase; DLD; LDHD; Probable D-lactate dehydrogenase, mitochondrial
Gene Aliases: 4733401P21Rik; Ac2-202; D8Bwg1320e; DLD; LDHD
UniProt ID: (Human) Q86WU2, (Mouse) Q7TNG8
Entrez Gene ID: (Human) 197257, (Rat) 307858, (Mouse) 52815
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support