Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MGPWGWKLR WTVALLLAAA GTAVGDRCER NEFQCQDGKC ISYKWVCDGS AECQDGSDES QETCLSVTCK SGDFSCGGRV NRCIPQFWRC DGQVDCDNGS DEQGCPPKTC SQDEFRCHDG KCISRQFVCD SDRDCLDGSD EASCPVLTCG PASFQCNSST CIPQLWACDN DPDCEDGSDE WPQRCRGLYV FQGDSSPCSA FEFHCLSGEC IHSSWRCDGG PDCKDKSDEE NCAVATCRPD EFQCSDGNCI HGSRQCDREY DCKDMSDEVG CVNVTLCEGP NKFKCHSGEC ITLDKVCNMA RDCRDWSDEP IKECGTNECL DNNGGCSHVC NDLKIGYECL CPDGFQLVAQ R (1-350 aa encoded by BC014514)
The low density lipoprotein receptor (LDL-R) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. Low density lipoprotein (LDL) is normally bound at the cell membrane and taken into the cell ending up in lysosomes where the protein is degraded and the cholesterol is made available for repression of microsomal enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG CoA) reductase, the rate-limiting step in cholesterol synthesis. At the same time, a reciprocal stimulation of cholesterol ester synthesis takes place. Mutations in the LDL-R gene cause the autosomal dominant disorder, familial hypercholesterolemia. Along with SCARB1, CLDN1, and the tetraspanin superfamily member CD81, LDL-R has been reported to be an entry factor for the Hepatitis C virus. At least three isoforms of LDL-R are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LDL receptor; ldl-r; Low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3
Gene Aliases: FH; FHC; LDLCQ2; LDLR
UniProt ID: (Human) P01130
Entrez Gene ID: (Human) 3949
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support