Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTPGRCCLAA DIQVETFIFT DGEDEALARH TGNVVITNCS AAHSRQALSC KMAVEYDRFI ESGRKWFCHV DDDNYVNLRA LLRLLASYPH TRDVYVGKPS LDRPIQAMER VSENKVRPVH FWFATGGAGF CISRGLALKM SPWASGGHFM NTAERIRLPD DCTIGYIVEA LLGVPLIRSG LFHSHLENLQ QVPTSELHEQ VTLSYGMFEN KRNAVHVKGP FSVEADPSRF RSIHCHLYPD TPWCPRTAIF
LFNG belongs to evolutionarily conserved glycosyltransferases that act in the Notch signaling pathway to define boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta-1,3-N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling. This protein is predicted to be a single-pass type II Golgi membrane protein but it may also be secreted and proteolytically processed like the related proteins in mouse and Drosophila (PMID: 9187150).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-N-acetylglucosaminyltransferase lunatic fringe; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases: LFNG; SCDO3
UniProt ID: (Human) Q8NES3
Entrez Gene ID: (Human) 3955
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support