Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ecalectin; Gal-9; Gal9; Galectin-9; Galectin9; HOM HD 21; lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; Tumor antigen HOM-HD-21; urate transporter/channel protein
Gene Aliases: HUAT; LGALS9; LGALS9A
UniProt ID: (Human) O00182
Entrez Gene ID: (Human) 3965
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support