Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
1 µg/mL of MA5-27652 was sufficient for detection of LGI1 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 60kDa.
This antibody was formerly sold as clone S283-7.
The leucine-rich, glioma inactivated gene 1 (LGI1) was first identified as a candidate tumor suppressor gene for glioma and may play a role in other cancers. LGI1 is a member of a family of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 serves as a ligand to ADAM22, a metalloprotease localized at the synapse. Mutations in LGI1 account for nearly half of autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Two isoforms of LGI1 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epitempin 1; Epitempin-1; leucine-rich glioma inactivated 1; Leucine-rich glioma-inactivated protein 1; leucine-rich repeat LGI family, member 1; leucine-rich, glioma inactivated 1; OTTHUMP00000020121; OTTHUMP00000020122
Gene Aliases: ADLTE; ADPAEF; ADPEAF; BB130740; EPITEMPIN; EPT; ETL1; IB1099; LGI1; UNQ775/PRO1569
UniProt ID: (Human) O95970, (Rat) Q8K4Y5, (Mouse) Q9JIA1
Entrez Gene ID: (Human) 9211, (Rat) 252892, (Mouse) 56839
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support