Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
G protein-coupled receptor GPR49 has been reported to be expressed in brain, skeletal muscle, placenta, and spinal cord. ESTs have been isolated from normal human brain (amygdala), embryo, and placenta libraries and from human germ cell, uterus, and brain libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This "turn-off" activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: follicle expressed hormone; G protein-coupled receptor 49; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; Leucine-rich repeat-containing G-protein coupled receptor 5; orphan G protein-coupled receptor HG38; Orphan G-protein coupled receptor FEX
Gene Aliases: FEX; GPR49; GPR67; GRP49; HG38; LGR5
UniProt ID: (Human) O75473, (Mouse) Q9Z1P4
Entrez Gene ID: (Human) 8549, (Mouse) 14160
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support