Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: SGVLYDSGAG GGTAIAGSVE AVARLKRSRL KVRFCTNESQ KSRAELVGQL QRLGFDISEQ EVTAPAPAAC QILKERGLRP YLLIHDGVRS EFDQIDTSNP NCVVIADAGE
Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 90%.
LHPP is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hLHPP; Phospholysine phosphohistidine inorganic pyrophosphate phosphatase
Gene Aliases: HDHD2B; LHPP
UniProt ID: (Human) Q9H008
Entrez Gene ID: (Human) 64077
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support