Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Signal-transducing molecule. May have a common pathway with IL6ST. The soluble form inhibits the biological activity of LIF by blocking its binding to receptors on target cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD118; CD118 antigen; D-factor/LIF receptor; FLJ98106; FLJ99923; leukemia inhibitor factor receptor alpha-chain; Leukemia inhibitory factor receptor; leukemia inhibitory factor receptor alpha chain; LIF R beta; LIF receptor; LIF-R; soluble differentiation-stimulating factor receptor
Gene Aliases: A230075M04Rik; AW061234; CD118; LIF; LIF-R; LIFR; SJS2; STWS; SWS
UniProt ID: (Human) P42702, (Rat) O70535, (Mouse) P42703
Entrez Gene ID: (Human) 3977, (Rat) 81680, (Mouse) 16880
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support