Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This antibody The antibody LIME-06 was raised against intracellular fragment corresponding to aa 141-295 of human LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed by T cells.
LIME (Lck-interacting molecule) is a 30 kDa double-palmitoylated protein with unusually basic cytoplasmic domain, expressed by T cells. After ligation of CD4 or CD8 T cell coreceptors, LIME is phosphorylated by Src-family kinases and associates with Lck and Fyn kinases and with their negative regulator Csk. Interestingly, Csk-mediated phosphorylation of C-terminal negative-regulatory tyrosine of LIME-associated Lck can result in increase of enzymatic activity compared with the total pool of Lck, thus, LIME serves as a positive regulator of TCR-dependent T cell signaling. However, under some circumstances, LIME may mediate inhibitory signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20406; Lck-interacting membrane protein; Lck-interacting molecule; Lck-interacting transmembrane adapter 1; lime 1; RP4-583P15.5
Gene Aliases: dJ583P15.4; LIME; LIME1; LP8067
UniProt ID: (Human) Q5JWJ2
Entrez Gene ID: (Human) 54923
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support