Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: KRLKKLSELT SRKAQPKATD LNSA
Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.
Adapter protein in a cytoplasmic complex linking beta- integrins to the actin cytoskeleton, bridges the complex to cell surface receptor tyrosine kinases and growth factor receptors. Plays a role in modulating cell spreading and migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ILK-binding protein; LIM and senescent cell antigen-like domains 2; LIM and senescent cell antigen-like-containing domain protein 2; LIM-like protein 2; LIM-type zinc finger domains 2; Particularly interesting new Cys-His protein 2; PINCH-2
Gene Aliases: LGMD2W; LIMS2; PINCH-2; PINCH2
UniProt ID: (Human) Q7Z4I7
Entrez Gene ID: (Human) 55679
Molecular Function:
cell junction protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support