Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MAALGEPVR LERDICRAIE LLEKLQRSGE VPPQKLQALQ RVLQSEFCNA VREVYEHVYE TVDISSSPEV RANATAKATV AAFAASEGHS HPRVVELPKT EEGLGFNIMG GKEQNSPIYI SRIIPGGIAD RHGGLKRGDQ LLSVNGVSVE GEHHEKAVEL LKAAQGKVKL VVRYTPKVLE EMESRFEKMR SAKRRQQT (1-197 aa encoded by BC053907)
Protein assembly at the post synaptic density (PSD) of neuronal synapses is mediated in part by protein interactions with PDZ (PSD-95/discs large/zona occludens-1) motifs. The Mammalian LIN-7 (MALS) proteins are a family of small synaptic proteins that are the mammalian homologs of LIN-7, a C. elegans protein essential for vulval development. MALS proteins are also known as Veli (Vertebrate Lin-7) proteins. MALS-3 mRNA is most abundant in kidney, followed by brain, liver, thymus and heart. MALS-3 is undetectable in spleen. In cultured cortical neurons, MALS proteins are clustered together with PSD-95 and NMDA-type glutamate receptors. MALS proteins are suggested to have roles in regulating the recruitment of neurotransmitter receptors to the PSD and in the assembly of proteins involved in synaptic vesicle exocytosis and synaptic junctions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11215; lin 7 homolog C; lin-7 homolog C; LIN-7 protein 3; lin-7-C; Lin-7C; MALS-3; Mammalian lin-seven protein 3; mLin7C; Protein lin-7 homolog C; Veli-3; veli-3 protein; vertebrate homolog of C. elegans Lin-7 type 3; Vertebrate lin-7 homolog 3
Gene Aliases: 9130007B12Rik; AI303698; AU019331; AW125731; D2Ertd520e; LIN-7-C; LIN-7C; LIN7C; MALS-3; MALS3; VELI3
UniProt ID: (Human) Q9NUP9, (Rat) Q792I0, (Mouse) O88952
Entrez Gene ID: (Human) 55327, (Rat) 60442, (Mouse) 22343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support