Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MHRGGQPLK KRRGSFKMAE LDQLPDESSS AKALVSLKEG SLSNTWNEKY SSLQKTPVWK GRNTSSAVEM PFRNSKRSRL FSDEDDRQIN TRSPKRNQRV AMVPQKFTAT MSTPDKKASQ KIGFRLRNLL KLPKAHKWCI YEWFYSNIDK PLFEGDNDFC VCLKESFPNL KTRKLTRVEW GKIRRLMGKP RRCSSAFFEE ERSALKQKRQ KIRLLQQRKV ADVSQFKDLP DEIPLPLVIG TKVTARLRGV HDGLFTGQID AVDTLNATYR VTFDRTGLGT HTIPDYEVLS NEPHETM (1-296 aa encoded by BC045625)
LIN9 belongs to the lin-9 family and acts as a tumor suppressor. It inhibits DNA synthesis. Its ability to inhibit oncogenic transformation is mediated through its association with RB1. LIN9 plays a role in the expression of genes required for the G1/S transition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta subunit-associated regulator of apoptosis; HuLin-9; lin-9 DREAM MuvB core complex component; lin-9 homolog; pRB-associated protein; Protein lin-9 homolog; rb related pathway actor; TUDOR gene similar 1 protein; TUDOR gene similar protein; Type I interferon receptor beta chain-associated protein
Gene Aliases: 2700022J23Rik; BARA; BARPsv; Lin-9; LIN9; mLin-9; TGS; TGS1; TGS2
UniProt ID: (Human) Q5TKA1
Entrez Gene ID: (Human) 286826, (Rat) 689523, (Mouse) 72568
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support