Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VVVVDWNRG ATTLIYTHAS SKTRKVAMVL KEFIDQMLAE GASLDDIYMI GVSLGAHISG FVGEMYDGWL GRITGLDPAG PLFNGKPHQD RLDPSDAQFV DVIHSDTDAL GYKEPLGNID FYPNGGLDQP GCPKTILGGF QYFKCDHQRS VYLYLSSLRE SCTITAYPCD SYQDYRNGKC VSCGTSQKES CPLLGYYADN WKDHLRGKDP PMTKAFFDTA EESPFCMYHY FVDIITWNKN VRRGDITIKL RDKAGNTTES KINHEPTTFQ KYHQVSLLAR FNQDLDKVAA ISLMFSTGSL IGPRYKLRIL RMKLRSLAHP ERPQLCRYDL VLMENVETVF QPILCPELQL (103-451 aa encoded by BC064941)
This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lipase H; Lipase member H; lipase, member H; LIPH; LPD lipase-related protein; Membrane-associated phosphatidic acid-selective phospholipase A1-alpha; membrane-bound phosphatidic acid-selective phospholipase A1; mPA-PLA1 alpha; phospholipase A(1); Phospholipase A1 member B
Gene Aliases: AH; ARWH2; C130037N08Rik; D16Wsu119e; H06; HYPT7; LAH2; LIPH; lpd1; lpd2; LPDLR; mPA-PLA1; MPAPLA1; P3; PLA1B
UniProt ID: (Human) Q8WWY8, (Mouse) Q8CIV3, (Rat) Q32PY2
Entrez Gene ID: (Human) 200879, (Mouse) 239759, (Rat) 681694
Molecular Function:
lipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support