Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is human brain tissue lysate.
PA5-21061 can be used with blocking peptide PEP-1175.
LMBRD1, also known as NESI (nuclear export signal-interacting protein, is a lysosomal membrane protein that is thought be involved in the transport and metabolism of cobalamin. LMBRD1 was initially identified as interacting with the large form of the hepatitis delta antigen and may be required for the nucleocytoplasmic shuttling of the hepatitis delta virus. Mutations in this gene are associated with the vitamin B12 metabolism disorder termed, homocystinuria-megaloblastic anemia complementation type F (cblF).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HDAg-L-interacting protein NESI; hepatitis delta antigen-L interacting protein; liver regeneration p-53 related protein; LMBD1; LMBR1 domain-containing protein 1; Lysosomal cobalamin transport escort protein LMBD1; Nuclear export signal-interacting protein; probable lysosomal cobalamin transporter; Protein N90b
Gene Aliases: 0910001K20Rik; AV347960; BM-021; C6orf209; CD001; LMBD1; LMBRD1; MAHCF; MSTP044; NESI
UniProt ID: (Human) Q9NUN5, (Mouse) Q3U696, (Rat) Q6AZ61
Entrez Gene ID: (Human) 55788, (Mouse) 68421, (Rat) 246046
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support