Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LGDIGTSCYT KSGMILCRND YIRLFGNSGA CSACGQSIPA SELVMRAQGN VYHLKCFTCS TCRNRLVPGD RFHYINGSLF CEHDRPTALI NGHLNSLQSN PLLPDQKVC
LMO4 encodes a cysteine-rich protein that contains two LIM domains but lacks a DNA-binding homeodomain. The encoded protein may play a role as a transcriptional regulator or as an oncogene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Breast tumor autoantigen; LIM domain only protein 4; LIM domain transcription factor LMO4; LIM-only 4 protein; LMO-4
Gene Aliases: LMO4
UniProt ID: (Human) O00158
Entrez Gene ID: (Human) 8543
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support