Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEELEKELDV VDPDGSVPVG LRQRNQTEKQ STGVYNREAM LNFCEKETKK LMQREMSMDE SKQVETKTDA KNGEERGRDA SKKALGPRRD SDLGKEPKRG GLKKSFSRDR DEAGGKSGEK PKEEKIIRGI DKGRVRAAVD KKEAGKDGRG EERAVATKKE EEKKGSDRNT GLSRDKDKKR EEMKEVAKKE DDEKVKGGAP AAPPPPPPPL APPLIMENLK NSLSPATQRK MGDKVLPAQE KNSRDQLLAA IRSSNLKQLK KVEVPKLLQ
The leiomodin 1 protein (LMOD1) has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves' disease and thyroid-associated ophthalmopathy. The leiomodin 1 protein has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves' disease and thyroid-associated ophthalmopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 64 kDa autoantigen 1D; 64 kDa autoantigen 1D3; 64 kDa autoantigen D1; leimodin 1 (smooth muscle); leiomodin 1 (smooth muscle); Leiomodin, muscle form; Leiomodin-1; SM-Lmod; Smooth muscle leiomodin; thyroid and eye muscle autoantigen D1 (64kD); Thyroid-associated ophthalmopathy autoantigen
Gene Aliases: 1D; 64kD; D1; LMOD1; SM-LMOD; SMLMOD
UniProt ID: (Human) P29536
Entrez Gene ID: (Human) 25802
Molecular Function: actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support