Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGTTSDEMVS VEQTSSSSLN PLCFECGQQH WTRENHLYNY QNEVDDDLVC HICLQPLLQP LDTPCGHTFC YKCLRNFLQE KDFCPLDRKR LHFKLCKKSS
LNX2 (Ligand of Numb Protein X 2) protein is encoded by the LNX2 gene located on chromosome 13 in humans and is known as PDZRN1 (PDZ domain-containing RING finger 1). It is through its PDZ domains that LNX2 forms oligomers and interacts with some of its target proteins. LNX2 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus modulates the Notch signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ligand of Numb protein X 2; Numb-binding protein 2; PDZ domain containing ring finger 1; PDZ domain-containing RING finger protein 1
Gene Aliases: LNX2; PDZRN1
UniProt ID: (Human) Q8N448
Entrez Gene ID: (Human) 222484
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support