Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSGNYQQS EALSKPTFSE EQASALVESV FGLKVSKVRP LPSYDDQNFH VYVSKTKDGP TEYVLKISNT KASKNPDLIE VQNHIIMFLK AAGFPTASVC HTKGDNTASL VSVDSGSEIK SYLVRLLTYL PGRPIAELPV SPQLLYEIGK LAAKLDKTLQ RFHHPKLSSL HRENFIWNLK NVPLLEKYLY ALGQNRNREI VEHVIHLFKE EVMTKLSHFR ECEYSPN (1-226 aa encoded by BC132753)
Catalyzes the GTP-dependent phosphorylation of 5-hydroxy-L-lysine. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-hydroxy-L-lysine kinase; 5-hydroxylysine kinase; aminoglycoside phosphotransferase domain containing 1; Aminoglycoside phosphotransferase domain-containing protein 1; Hydroxylysine kinase
Gene Aliases: AGPHD1; C630028N24Rik; HYKK
UniProt ID: (Human) A2RU49, (Mouse) Q5U5V2
Entrez Gene ID: (Human) 123688, (Mouse) 235386
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support