Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDSVVFEDVA VDFTLEEWAL LDSAQRDLYR DVMLETFRNL ASVDDGTQFK ANGSVSLQDM YGQEKSKEQT IPNFTGNNSC AYTLEKNCEG YGTEDHHKNL RNHMVDRFCT HNEGNQYGEA IHQMPDLTLH KKVSAGEKPY ECTKCRTVFT HLSSLKRHVK SHCGRKAPPG EECKQACICP SHLHSHGRTD TEEKPYKCQA CGQTFQHPRY LSHHVKTHTA EKTYKCEQCR MAFNGFASFT RHVRTHTKDR PYKCQECGRA FIYPSTFQRH MTTHTGEKPY KCQHCGKAFT YPQAFQRHEK THTGEKPYEC KQCGKTFSWS ETLRVHMRIH TGDKLYKCEH CGKAFTSSRS FQGHLRTHTG EKPYECKQCG KAFTWSSTFR EHVRIHTQEQ LYKCEQCGKA FTSSRSFRGH LRTHTGEKPY ECKQCGKTFT WSSTFREHVR IHTQEQLHKC EHCGKAFTSS RAFQGHLRMH TGEKPYECKQ CGKTFTWSST LHNHVRMHTG EKPHKCKQCG MSFKWHSSFR NHLRMHTGQK SHECQSYSKA FSCQVILSKT SESTH
The function of ZNF57 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 424; Zinc finger protein 57
Gene Aliases: ZNF424; ZNF57
UniProt ID: (Human) Q68EA5
Entrez Gene ID: (Human) 126295
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support