Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEQTKLNTIL CQTFVFCYLW SLGGNLTENY YDSFDTFIRT QFDDNPDARL PNSGDLWSIH MDFDTKRLDP WERIIPTFKY NRDVPFFEML VPTTDTVRYG YLMEKLLAVK HSVLFTGITG VGKSVIAKGL LNKIQESAGY VPVYLNFSAQ TSSARTQEII ESKLERKRKN ILGAPGNKRI VIFVDDLNMP RLDRYGSQPP IELLRQYQDF GGFYDRNKLF WKEIQDVTII SACAPPGGGR NPVTPRFIRH FSMLCLPMPS EHSLKQIFQA ILNGFLSDFP PAVKQTASSI VEASVEIYNK MSVDLLPTPA KSHYVFNLRD LSKCVQDKHF GIAIDLEYFL NKPIIFGDFI KFGADKADRI YDDMPDIEKT ANVLQDYLDD YNLTNPKEVK LVFFQDAIEH VSRIARMIRQ ERGNALLVGV GGTGKQSLTR LAAHICGYKC LQIELSRGYN YDSFHEDLRK LYKMAGVEDK NMVFLFTDTQ IVVEEFLEDI NNILNSGEVP NLFEKDELEQ VLAATRPRAK EVGISEGNRD EVGCARVVMW LYHKNVSSPF I
DNAH6 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonemal beta dynein heavy chain 6; Ciliary dynein heavy chain 6; Dynein axonemal heavy chain 6; dynein heavy chain-like 1; dynein, axonemal, heavy polypeptide 6
Gene Aliases: DNAH6; DNAHC6; DNHL1; HL-2; HL2; KIAA1697
UniProt ID: (Human) Q9C0G6
Entrez Gene ID: (Human) 1768
Molecular Function: microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support