Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWPFPSRSLF PPPTQAWLQT VSSDPEAQGW GAWNETKEIL GPEGGEGKEE KEEEEDAEED QDGDAGFLLS LLEQENLAEC PLPDQELEAI KMKVCAMEQA EGTPRPPGVQ QQAEEEEGTA AGQLLSPETV GCPLSGTPEE KVEADHRSVY VGNVDYGGSA EELEAHFSRC GEVHRVTILC DKFSGHPKGY AYIEFATKGS VQAAVELDQS LFRGRVIKVL PKRTNFPGIS STDRGGLRGH PGSRGAPFPH SGLQGRPRLR PQGQNRTPAA LTPLWARPLP WVVVSHVIK
Binds the poly(A) tail of mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: embryonic poly(A) binding protein 2; Embryonic poly(A)-binding protein 2; Embryonic poly(A)-binding protein type II; Embryonic polyadenylate-binding protein 2; ePABP-2; poly(A) binding protein, nuclear 1-like (cytoplasmic); Poly(A)-binding protein nuclear-like 1
Gene Aliases: EPABP2; PABPN1L; PABPNL1
UniProt ID: (Human) A6NDY0
Entrez Gene ID: (Human) 390748
Molecular Function: translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support