Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: mouse brain.
Predicted reactivity: Mouse (96%), Zebrafish (87%), Xenopus laevis (92%), Chicken (93%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ATP-dependent serine protease that mediates the selective degradation of misfolded and unassembled polypeptides in the peroxisomal matrix. Necessary for type 2 peroxisome targeting signal (PTS2)-containing protein processing and facilitates peroxisome matrix protein import.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lon peptidase 2; Lon protease 2; Lon protease homolog 2, peroxisomal; Lon protease-like protein 2; LONP2; MGC4840; Peroxisomal Lon protease; peroxisomal Lon protease homolog 2; peroxisomal LON protease like; pLon
Gene Aliases: 1300002A08Rik; AU015403; LONP; LONP2; LONPL
UniProt ID: (Human) Q86WA8, (Mouse) Q9DBN5
Entrez Gene ID: (Human) 83752, (Mouse) 66887
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support