Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PCAT1 acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to hosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase; 1-alkenylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; Acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholin; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; Acyltransferase-like 2; aytl2; LPC acyltransferase 1; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; Lysophosphatidylcholine acyltransferase 1; mLPCAT1; Phosphonoformate immuno-associated protein 3; regulated by phosphonoformate
Gene Aliases: 2900035H07Rik; AGPAT10; AGPAT9; AYTL2; BB137372; BC005662; C87117; LPCAT; LPCAT1; PFAAP3; rd11; RGD1311599; si:dkey-261i16.4; zgc:158232
UniProt ID: (Human) Q8NF37, (Mouse) Q3TFD2, (Rat) Q1HAQ0, (Zebrafish) Q1LWG4
Entrez Gene ID: (Dog) 488066, (Human) 79888, (Pig) 100217397, (Chicken) 420974, (Mouse) 210992, (Rat) 361467, (Zebrafish) 555969
Molecular Function: acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support