Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MSQGSPGDWA PLDPTPGPPA SPNPFVHELH LSRLQRVKFC LLGALLAPIR VLLAFIVLFL LWPFAWLQVA GLSEEQLQEP ITGWRKTVCH NGVLGLSRLL FFLLGFLRIR VRGQRASRLQ APVLVAAPHS TFFDPIVLLP CDLPKVVSRA ENLSVPVIGA LLRFNQAILV SRHDPASRRR VVEEVRRRAT SGGKWPQVLF FPEGTCSNKK ALLKFKPGAF IAGVPVQPVL IRYPNSLDTT SWAWRGPGVL KVLWLTASQP CSIVDVEFLP VYHPSPEESR DPTLYANNVQ RVMAQALGIP ATECEFVGSL PVIVVGRLKV ALEPQLWELG KVLRKAGLSA GYVDAGAEPG RSRMISQEEF ARQLQLSDPQ TVAGAFGYFQ QDTKGLVDFR DVALALAALD GGRSLEELTR LAFELFAEEQ AEGPNRLLYK DGFSTILHLL LGSPHPAATA LHAELCQAGS SQGLSLCQFQ NFSLHDPLYG KLFSTYLRPP HTSRGTSQTP NASSPGNPTA LANGTVQAPK QKGD
Members of the 1-acylglycerol-3-phosphate O-acyltransferase family, such as AGPAT7, catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA), a precursor in the biosynthesis of all glycerolipids. Both LPA and PA are involved in signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase 7; 1-acylglycerol-3-phosphate O-acyltransferase 7 (lysophosphatidic acid acyltransferase, eta); 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase; 1-AGP acyltransferase 7; 1-AGPAT 7; 1-alkenylglycerophosphoethanolamine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; acyl-CoA:lysophosphatidylethanolamine acyltransferase 2; acyltransferase like 3; Acyltransferase-like 3; Lysophosphatidylcholine acyltransferase 4; Lysophosphatidylethanolamine acyltransferase 2; Lysophospholipid acyltransferase LPCAT4; Plasmalogen synthase; PLSC domain containing protein
Gene Aliases: AGPAT7; AYTL3; LPAAT-eta; LPCAT4; LPEAT2
UniProt ID: (Human) Q643R3
Entrez Gene ID: (Human) 254531
Molecular Function: acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support