Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: DQQLLLLKKH LENNYRSRDR KWIVLFPEGG FLRKRRETSQ AFAKKNNLPF
LPGAT1 is a lysophoshatidylglycerol (LPG) specific acyltransferase that recognizes various acyl-CoAs and LPGs as substrates but demonstrates a clear preference for long chain saturated fatty acyl-CoAs and oleoyl-CoA as acyl donors. It prefers oleoyl-LPG over palmitoyl-LPG as an acyl receptor and oleoyl-CoA over lauroyl-CoA as an acyl donor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-acylglycerophosphocholine O-acyltransferase; Acyl-CoA:lysophosphatidylglycerol acyltransferase 1; Acyl-CoA:monoacylglycerol acyltransferase LPGAT1; family with sequence similarity 34, member A; LPLAT7; Lysophospholipid acyltransferase 7; Stearoyl-CoA:1-lyso-2-acyl-PE acyltransferase
Gene Aliases: FAM34A; FAM34A1; KIAA0205; LPGAT1; NET8
UniProt ID: (Human) Q92604
Entrez Gene ID: (Human) 9926
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support