Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Latrophilin family of G protein-coupled receptors consists of three members, latrophilin-1, 2 and 3, each of which displays distinct tissue distribution and function. Latrophilin-1, the most characterized member of this family, acts as a receptor for a-latrotoxin, a component of venom from the black widow spider. Binding of a-latrotoxin to latrophilin-1 triggers synaptic vesicle exocytosis via both Ca2+-dependent and -independent mechanisms, resulting in vesicle depletion. Latrophilin-1 is abundant in brain and present in endocrine cells. Latrophilin-3 is also brain-specific, whereas latrophilin-2 expression is ubiquitous.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G protein-coupled receptor L2; Calcium-independent alpha-latrotoxin receptor 2; calcium-independent alpha-latrotoxin receptor homolog 2; CIRL-2; Cl2aa; latrophilin 1; latrophilin 2; Latrophilin homolog 1; Latrophilin-2; Lectomedin-1
Gene Aliases: ADGRL2; AI450192; CIRL2; CL2; Gm619; KIAA0786; LEC1; LPHH1; LPHN2; mKIAA0786
UniProt ID: (Human) O95490
Entrez Gene ID: (Human) 23266, (Mouse) 99633
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support