Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This Lactoperoxidase antibody is useful for Western blot analysis where a band is seen at ~100 kDa.
Caution, do not use milk for the blocking or dilution buffers.
Suggested positive control: MDA-MB231 cell lysate.
This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: airway lactoperoxidase; EC 1.11.17 antibody; Lactoperoxidase; lactoperoxydase; LPO; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX antibody; SPO; SPO antibody
Gene Aliases: 5830499B15Rik; LPO; SAPX; SPO
UniProt ID: (Goat) A0A452E9Y6, (Human) P22079, (Bovine) P80025, (Mouse) Q5SW46
Entrez Gene ID: (Goat) 100860761, (Human) 4025, (Sheep) 443004, (Bovine) 280844, (Mouse) 76113
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support