Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 1.11.17 antibody; Lactoperoxidase; LPO; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX antibody; SPO; SPO antibody
Gene Aliases: LPO; SAPX; SPO
UniProt ID: (Human) P22079
Entrez Gene ID: (Human) 4025
Molecular Function: peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support