Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VHNFQSRPPS GRRLSPWEDL GQAPTMDSPL EKNPRSAGRI RHRHGSPHPS RRTA
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
LPPR2 is a multi-pass membrane protein and belongs to the PA-phosphatase related phosphoesterase family. LPPR2 shows dynamic expression regulation during brain development and neuronal excitation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive phospholipid phosphatase PLPPR2; Lipid phosphate phosphatase-related protein type 2; Phospholipid phosphatase-related protein type 2; plasticity related gene 4; Plasticity-related gene 4 protein; PRG-4
Gene Aliases: LPPR2; PLPPR2; PRG4
UniProt ID: (Human) Q96GM1
Entrez Gene ID: (Human) 64748
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support