Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects endogenous protein at a molecular weight of 25 kDa (monomer) and 50 kDa (dimer).
Purity is >95% by SDS-PAGE.
Lecithin retinol acyltransferase (LRAT) is a membrane bound enzyme that catalyzes the transfer of an acyl group from the sn-1 position of lecithin to vitamin A which generates all-trans-retinyl esters (tREs) in the liver, some extrahepatic tissues, such as the lung, and retinal pigmented epithelium. LRAT can also exchange palmitoyl groups between RPE65, a tRE binding protein essential for vision, and tREs, which is important for the operation of the visual pathway. LRAT is essential for the dietary mobilization, transport, and storage of vitamin A as well as the synthesis of the visual pigment chromophore. LRAT monomers interact in membranes to form homodimers through disulfide bond formation. A loss of LRAT correlates with an early onset severe retinal dystrophy and severe retinyl ester deprivation, while a reduction in LRAT expression may be associated with invasive bladder cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lecithin retinol acyltransferase; lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase); Phosphatidylcholine--retinol O-acyltransferase; Phosphatidylcholine-retinol-O-acyltransferase
Gene Aliases: 1300010A18Rik; AI449251; LCA14; LRAT
UniProt ID: (Human) O95237, (Mouse) Q9JI60, (Rat) Q9JI61
Entrez Gene ID: (Human) 9227, (Mouse) 79235, (Rat) 64047
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support