Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: YFKPPFGPYP EKSGKDSLVS MKCSLFRFCP WSKELPFQPP EGSISSHLGS GASDSETEET RKALPIQSFS HEKESHQHRQ HSVPVISRPG SNVKPTLPPI PQGRR
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich repeat and guanylate kinase domain-containing protein; leucine-rich repeats and guanylate kinase domain containing
Gene Aliases: CFAP246; LRGUK
UniProt ID: (Human) Q96M69
Entrez Gene ID: (Human) 136332
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support