Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: YQTRKKSEEY SVTNTDETVV PPDVPSYLSS QGTLSDRQET VVRTEGGPQA NGHIESNGVC PRDASHFPEP DTHSVACRQP KLCAGSAYHK EPWKAMEKAE GTPGPHKMEH GGRVVCSDCN TEVDCYSRGQ AFHPQPVSRD SAQPSAPNGP EPGGSDQEHS PHHQCSRTAA GSCPECQGSL YPSNHDRMLT AVKKKPMASL DGKGDSSWTL ARLYHPDSTE LQPASSLTSG SPERAEAQYL LVSNGHLPKA CDASPESTPL TGQLPGKQRV PLLLAPKS; Positive Samples: Raji, HeLa; Cellular Location: Membrane, Single-pass type I membrane protein
LRIG1 protein Q96JA1 is coded by a gene on a chromosome band 3p14.3, this region is known to be deleted in various human cancers. LRIG1 is considered to be a tumor suppressor gene in humans. LRIG1 is an integral cell-surface membrane protein that is expressed by specific cells in various human tissues and that its 143-kDa form might be cleaved into 111-kDa and 32-kDa fragments. The LRIG1 protein may inhibit the growth of tumors of glial cells and the down-regulation of the LRIG1 gene may be involved in the development and progression of the tumor.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp586O1624; leucine-rich repeat protein LRIG1; leucine-rich repeats and immunoglobulin like domains 1; leucine-rich repeats and immunoglobulin-like domains 1; Leucine-rich repeats and immunoglobulin-like domains protein 1; LIG-1; ortholog of mouse integral membrane glycoprotein LIG-1
Gene Aliases: LIG-1; LIG1; LRIG1
UniProt ID: (Human) Q96JA1
Entrez Gene ID: (Human) 26018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support