Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: KVCKLQCKSE PFWEDDLAKE TYIQFETLFP RSQSVGELWT RSHRDDSEKL LLCSRSSVES QVTFKSEGSR PEYYC
This gene encodes a protein that has a fibronectin type III domain and a C-terminal transmembrane domain, as well as a leucine-rich repeat domain and immunoglobulin-like domain near the N-terminus. The encoded protein may regulate fibroblast growth factor receptors and affect the modification of these receptors, which are glycosylated differently in the Golgi and endoplasmic reticulum. Mutations in this gene are associated with congenital stationary night blindness, type 1F.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibronectin type III, immunoglobulin and leucine rich repeat domains 4; leucine-rich repeat, Ig-like and transmembrane domains 3; leucine-rich repeat, immunoglobulin-like and transmembrane domains 3; Leucine-rich repeat, immunoglobulin-like domain and transmembrane domain-containing protein 3
Gene Aliases: CSNB1F; FIGLER4; LRIT3
UniProt ID: (Human) Q3SXY7
Entrez Gene ID: (Human) 345193
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support