Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: A549, HeLa, HepG2, HCT116.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a deadenylase that functions as the catalytic subunit of the polyadenylate binding protein dependent poly(A) nuclease complex. The encoded protein is a magnesium dependent 3' to 5' exoribonuclease that is involved in the degradation of cytoplasmic mRNAs. Alternate splicing results in multiple transcript variants.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 130 kDa leucine-rich protein; GP130; leucine-rich pentatricopeptide repeat containing; Leucine-rich PPR motif-containing protein, mitochondrial; leucine-rich PPR-motif containing; LRP 130; mitochondrial leucine-rich PPR motif-containing protein
Gene Aliases: CLONE-23970; GP130; LRP130; LRPPRC; LSFC
UniProt ID: (Human) P42704
Entrez Gene ID: (Human) 10128
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support