Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
For maximum recovery of product, centrifuge the vial prior to removing the cap.
This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Corin; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; Low-density lipoprotein receptor-related protein 4; LRP-4; Multiple epidermal growth factor-like domains 7
Gene Aliases: 6430526J12Rik; CLSS; CMS17; D230026E03; KIAA0816; LRP-4; LRP10; LRP4; mdig; MEGF7; SOST2
UniProt ID: (Human) O75096, (Mouse) Q8VI56
Entrez Gene ID: (Human) 4038, (Rabbit) 100346254, (Rat) 83469, (Mouse) 228357
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support