Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: U-87MG, Mouse testis, Rat testis
Immunogen sequence: PVYRKTTEEE DEDELHIGRT AQIGHVYPAA ISSFDRPLWA EPCLGETREP EDPAPALKEL FVLPGEPRSQ LHQLPKNPLS ELPVVKSKRV ALSLEDDGLP
Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ApoE receptor 2; Apolipoprotein E receptor 2; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; Low-density lipoprotein receptor-related protein 8; LRP-8
Gene Aliases: 4932703M08Rik; AA921429; AI848122; APOER2; HSZ75190; Lr8b; LRP-8; LRP8; MCI1
UniProt ID: (Human) Q14114
Entrez Gene ID: (Human) 7804, (Rat) 362558, (Mouse) 16975
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support