Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: QTCPSVCSC SNQFSKVICV RKNLREVPDG ISTNTRLLNL HENQIQIIKV NSFKHLRHLE ILQLSRNHIR TIEIGAFNGL ANLNTLELFD NRLTTIPNGA FVYLSKLKEL WLRNNPIESI PSYAFNRIPS LRRLDLGELK RLSYISEGAF EGLSNLRYLN LAMCNLREIP NLTPLIKLDE LDLSGNHLSA IRPGSFQGLM HLQKLWMIQS QIQVIERNAF DNLQSLVEIN LAHNNLTLLP HDLFTPLHHL ERIHLHHNPW NCNCDILWLS WWIKDMAPSN TACCARCNTP PNLKGRYIGE LDQNYFTC (45-351 aa encoded by BC041374 )
Leucine-rich repeat-containing 4C (LRRC4C), also known as Netrin-G ligand 2 (NGL-2), is a gene highlighted for its expression in the nervous system, particularly the brain. It is a member of the leucine-rich repeat superfamily, which plays a significant role in cell adhesion and signaling pathways critical for synaptic connectivity and neuronal development. LRRC4C is involved in neural processes such as axon guidance and synapse formation, aiding in the establishment of functional neural circuits. Additionally, research has suggested that LRRC4C may function as a tumor suppressor, particularly in gliomas, indicating its potential role in neuro-oncological disorders. Variations or deletions in this gene have been linked to neurodevelopmental disorders, demonstrating its importance in normal brain function and development. Furthermore, LRRC4C's expression patterns and associations with cancer also suggest its potential as a prognostic marker in clinical settings.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich repeat-containing protein 4C; Netrin-G1 ligand; NGL-1
Gene Aliases: 6430556C10Rik; KIAA1580; LRRC4C; NGL-1; NGL1; RGD1311013; UNQ292/PRO331
UniProt ID: (Human) Q9HCJ2, (Mouse) Q8C031
Entrez Gene ID: (Human) 57689, (Mouse) 241568, (Rat) 311236
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support