Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MVSSNFWFKY PKTCSKVEHF VSILGKCFES PWTTKALSET ACEDSEENKQ RITGAQTLPK HVSTSSDEGS PSASTPMINK TGFKFSAEKP VIEVPSMTIL DKKDGEQAKA LFEKVRKFRA HVEDSDLIYK LYVVQTASPF PNQ
Component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8D constitutes a pore-forming subunit or whether it is closely associated with the pore.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsLRRC8D; leucine rich repeat containing 5; leucine rich repeat containing 8 family; leucine rich repeat containing 8 family, member D; leucine-rich repeat containing 8 family member D; leucine-rich repeat-containing 5; Leucine-rich repeat-containing protein 5; Leucine-rich repeat-containing protein 8D; Volume-regulated anion channel subunit LRRC8D
Gene Aliases: LRRC5; LRRC8D; UNQ213/PRO239
UniProt ID: (Human) Q7L1W4
Entrez Gene ID: (Human) 55144
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support