Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: QNLAITANRI ETLPPELFQC RKLRALHLGN NVLQSLPSRV GELTNLTQIE LRGNRLECLP VELGECPLLK RSGLVVEEDL FNTLPPEVKE RLWRADKEQA
Essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8A constitutes a pore-forming subunit or whether it is closely associated with the pore and mediates channel properties such as ion selectivity. Involved in B-cell development: required for the pro-B cell to pre-B cell transition. Also required for T-cell development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsLRRC8A; leucine rich repeat containing 8 family, member A; leucine-rich repeat containing 8 family member A; Leucine-rich repeat-containing protein 8A; Swelling protein 1; Volume-regulated anion channel subunit LRRC8A
Gene Aliases: AGM5; KIAA1437; LRRC8; LRRC8A; SWELL1; UNQ221/PRO247
UniProt ID: (Human) Q8IWT6
Entrez Gene ID: (Human) 56262
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support