Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLKENNLKSI EEIVSFQHLR KLTVLKLWHN SITYIPEHIK KLTSLERLSF SHNKIEVLPS HLFLCNKIRY LDLSYNDIRF IPPEIGVLQS LQYFSITCNK VESLPDELYF CKKLKTLKIG KNSLSVLSPK IGNLLFLSYL DVKGNHFEIL PPELGDCRAL KRAGLVVEDA LFETLPSDVR EQMKTE
LRRC8C is a protein coding gene. Non-essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and can also conduct organic osmolytes like taurine. Plays a redundant role in the efflux of amino acids, such as aspartate and glutamate, in response to osmotic stress. Channel activity requires LRRC8A plus at least one other family member (LRRC8B, LRRC8C, LRRC8D or LRRC8E); channel characteristics depend on the precise subunit composition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Factor for adipocyte differentiation 158; hypothetical protein AD158; leucine rich repeat containing 8 family, member C; leucine rich repeat containing 8C; leucine-rich repeat containing 8 family member C; Leucine-rich repeat-containing protein 8C; Volume-regulated anion channel subunit LRRC8C
Gene Aliases: AD158; FAD158; LRRC8C
UniProt ID: (Human) Q8TDW0
Entrez Gene ID: (Human) 84230
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support