Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: RVQETHPYHI LLSWVTPPNT VSTNLTWSSA SSLRGQGATA LARLPRGTHS YNITRLLQAT EYWACLQVAF ADAHTQLACV WARTKEATSC HRALGDRPG
The protein encoded by this gene belongs to the leucine-rich repeat superfamily. This gene was found to be amplified and overexpressed in malignant gliomas. The encoded protein has homology with other proteins that function as cell-adhesion molecules or as signal transduction receptors and is a candidate for the target gene in the 1q32.1 amplicon in malignant gliomas. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibronectin type III, immunoglobulin and leucine rich repeat domain 7; Glioma amplified on chromosome 1 protein; leucine rich and ankyrin repeats 1; leucine rich repeat neuronal 5; Leucine-rich repeat neuronal protein 2; Leucine-rich repeat neuronal protein 5
Gene Aliases: FIGLER7; GAC1; LRANK1; LRRN2; LRRN5; UNQ256/PRO293
UniProt ID: (Human) O75325
Entrez Gene ID: (Human) 10446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support