Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPLFFRKRKP SEEARKRLEY QMCLAKEAGA DDILDISKCE LSEIPFGAFA TCKVLQKKVL IVHTNHLTSL LPKSCSLLSL ATIKVLDLHD NQLTALPDDL GQLTALQVLN VERNQLMQLP RSIGNLTQLQ TLNVKDNKLK ELPDTVGELR SLRTLNISGN EIQRLPQMLA HVRTLEMLSL DASAMVYPPR EVCGAGTAAI LQFLCKESGL EYYPPSQYLL PILEQDGIEN SRDSPDGPTD RFSREELEWQ NRFSDYEKRK EQKMLEKLEF ERRLELGQRE HTQLLQQSSS QKDEILQTVK EEQSRLEQGL SEHQRHLNAE RQRLQEQLKQ TEQNISSRIQ KLLQDNQRQK KSSEILKSLE NERIRMEQLM SITQEETESL RRRDVASAMQ QMLTESCKNR LIQMAYESQR QNLVQQACSS MAEMDERFQQ ILSWQQMDQN KAISQILQES AMQKAAFEAL QVKKDLMHRQ IRSQIKLIET ELLQLTQLEL KRKSLDTESL QEMISEQRWA LSSLLQQLLK EKQQREEELR EILTELEAKS ETRQENYWLI QYQRLLNQKP LSLKLQEEGM ERQLVALLEE LSAEHYLPIF AHHRLSLDLL SQMSPGDLAK VGVSEAGLQH EILRRVQELL DAARIQPELK PPMGEVVTPT APQEPPESVR PSAPPAELEV QASECVVCLE REAQMIFLNC GHVCCCQQCC QPLRTCPLCR QDIAQRLRIY HSS
LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase LRSAM1; hTAL; Leucine-rich repeat and sterile alpha motif-containing protein 1; RING-type E3 ubiquitin transferase LRSAM1; Tsg101-associated ligase
Gene Aliases: CMT2P; LRSAM1; RIFLE; TAL; UNQ6496/PRO21356
UniProt ID: (Human) Q6UWE0
Entrez Gene ID: (Human) 90678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support