Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
LSM14A, Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AlphaSNBP; family with sequence similarity 61, member A; hRAP55; hRAP55A; LSM14 homolog A; LSM14A mRNA processing body assembly factor; LSM14A, SCD6 homolog A; mRAP55A; Protein FAM61A; Protein LSM14 homolog A; Protein SCD6 homolog; Putative alpha-synuclein-binding protein; RNA-associated protein 55; RNA-associated protein 55A
Gene Aliases: 2700023B17Rik; AA407828; AU017544; C19orf13; FAM61A; LSM14A; RAP55; RAP55A; Tral
UniProt ID: (Human) Q8ND56, (Mouse) Q8K2F8
Entrez Gene ID: (Human) 26065, (Mouse) 67070
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support