Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAANATTNP SQLLPLELVD KCIGSRIHIV MKSDKEIVGT LLGFDDFVNM VLEDVTEFEI TPEGRRITKL DQILLNGNNI TMLVPGGEGP EV (1-91 aa encoded by BC005938)
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LSM5 homolog, U6 small nuclear RNA associated; LSM5 U6 small nuclear RNA and mRNA degradation associated; U6 snRNA-associated Sm-like protein LSm5
Gene Aliases: 2010208O10Rik; 2310034K10Rik; LSM5; YER146W
UniProt ID: (Human) Q9Y4Y9, (Mouse) P62322
Entrez Gene ID: (Human) 23658, (Mouse) 66373
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support